Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  50  60  70  80  90  98  99  100  101  102  103  104  105  106  107  108  109  110  111  112  113  114  115  116  117  118  119  120  121  122  123  124  125  126  127  130  140  150  160  170  180  190  200  210  300  400  500  600  700  800  900  1000  1100  1630 
3361 - 3390 of 48882 products displayed
  • POMC Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518368
  • Size 100µg
Applications
Immunohistochemistry (4-6 µg/ml), Western Blot (0.1-0.3 µg/ml)
Alternate names:
Corticotropin-lipotropin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518368 349.00
Check your price
  • Staphylococcus aureus Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA110709
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
S. aureus
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110709 349.00
Check your price
  • Sheep anti-Human IgA Secondary Antibody, Biotin conjugate for IF
  • Host Sheep
  • Target Species Human
  • Conjugate Biotin
  • Catalog # PA516923
  • Size 2mL
Applications
Immunofluorescence (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516923 109.00
Check your price
  • NFATC1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA141041
  • Size 100µg
Applications
Immunocytochemistry (1:10), Immunofluorescence (1:10), Western Blot (1-3 µg/ml)
Alternate names:
NFAT transcription complex cytosolic component
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141041 349.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, Alexa Fluor® 647 conjugate for IF, ICC and IHC
  • Host Goat
  • Target Species Chicken
  • Conjugate Alexa Fluor® 647
  • Catalog # A21449
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21449 169.00
Check your price
  • Mouse IgG1/Rat IgG1 Isotype Control, FITC, PE conjugate for Flow and Ctrl
  • Host Mixed
  • Target Species Not Applicable
  • Conjugate FITC , PE
  • Catalog # PA533183
  • Size 50TESTS
Applications
Control (Assay-Dependent), Flow Cytometry (Neat)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533183 375.00
Check your price
  • CCR8 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112539
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
TER1; GPR-CY6; Chemokine receptor-like 1; CMKBRL2; CC chemokine receptor CHEMR1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112539 325.00
Check your price
  • Tau Monoclonal Antibody for Western Blot, IF, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515108
  • Size 100µL
Applications
Immunofluorescence (1:800), Immunohistochemistry (Frozen) (1:800), Immunohistochemistry (Paraffin) (1:800), Western Blot (1:1000)
Alternate names:
Paired helical filament-tau; Neurofibrillary tangle protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515108 275.00
Check your price
  • PTPN14 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531285
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Protein-tyrosine phosphatase pez
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531285 349.00
Check your price
  • MGMT Monoclonal Antibody for Western Blot, IHC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 357000
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
O-6-methylguanine-DNA-alkyltransferase; 6-O-methylguanine-DNA methyltransferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
357000 304.00
Check your price
  • SMARCAL1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131918
  • Size 50µg
Applications
Western Blot (1:500)
Alternate names:
Sucrose nonfermenting protein 2-like 1; HepA-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131918 375.00
Check your price
  • Progesterone Receptor Monoclonal Antibody for Western Blot, IF, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Horse, Human, Pig
  • Conjugate Unconjugated
  • Catalog # MA112626
  • Size 100µg
Applications
Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin, Frozen) (1:25-1:50), Immunoprecipitation (2 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112626 475.00
Check your price
  • Goat anti-Human IgA Fc Secondary Antibody, FITC conjugate for Flow
  • Host Goat
  • Target Species Human
  • Conjugate FITC
  • Catalog # H14001
  • Size 2ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
H14001 213.00
Check your price
  • Galectin 3 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1940
  • Size 100µg
Applications
ELISA (Assay dependent), Immunocytochemistry (1:20-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (6 µg/ml), Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (1 µg/ml)
Alternate names:
Mac-2 antigen; Lectin L-29; Laminin-binding protein; IgE-binding protein; Galactose-specific lectin 3; Carbohydrate-binding protein 35; 35 kDa lectin; L-31; Galactoside-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1940 375.00
Check your price
  • GM-CSF Monoclonal Antibody for Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523699
  • Size 500µg
Applications
ELISA (0.5-2 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells)
Alternate names:
Sargramostim; Molgramostin; Colony-stimulating factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523699 349.00
Check your price
  • Calpain S1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531110
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Calpain regulatory subunit; Calcium-dependent protease small subunit 1; Calcium-dependent protease small subunit; Calcium-activated neutral proteinase small subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531110 349.00
Check your price
  • RGS5 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523231
  • Size 100µL
Applications
Western Blot (1:250-1:1000)
Alternate names:
RP11-430G6.2, MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523231 349.00
Check your price
  • IL-11 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA172508
  • Size 100µg
Applications
ELISA (1 µg/ml), Western Blot (1µg/ml)
Alternate names:
Adipogenesis inhibitory factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA172508 199.00
Check your price
  • Bovine Papilloma Virus Type 1 E2 Monoclonal Antibody for IF, ICC and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171536
  • Size 100ug
Applications
ELISA (0.2-1 ug/ml), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay dependent)
Alternate names:
BPV, BPV-1, BPV1-E2, BPV Protein E2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171536 325.00
Check your price
  • Phospho-PDX1 (Thr11) Polyclonal Antibody for DB
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513046
  • Size 400µL
Applications
Dot blot (1:500)
Alternate names:
Somatostatin-transactivating factor 1; Islet/duodenum homeobox-1; Insulin upstream factor 1; Insulin promoter factor 1; Glucose-sensitive factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513046 349.00
Check your price
  • HLA-DR Monoclonal Antibody, Pacific Orange™ conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Pacific Orange™
  • Catalog # MHLDR30
  • Size 0.5ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
MHC class II antigen DRB1*15; DW2.2/DR2.2; MHC class II antigen DRB1*16; MHC class II antigen DRB1*3; Clone P2-beta-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHLDR30 420.00
Check your price
  • ICAM-1 Monoclonal Antibody, APC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate APC
  • Catalog # A15396
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
MyD10; MALA-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15396 192.00
Check your price
  • CD9 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # A15830
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
p24; Tetraspanin-29; Motility-related protein; Leukocyte antigen MIC3; Cell growth-inhibiting gene 2 protein; 5H9 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15830 117.00
Check your price
  • Rabbit anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 488 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Goat
  • Conjugate DyLight® 488
  • Catalog # SA510078
  • Size 500µg
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510078 156.00
Check your price
  • CCL3 Polyclonal Antibody for Western Blot, IHC (F), ELISA and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546951
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunohistochemistry (Frozen) (5-15 µg/ml), Neutralization (0.05-0.2 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
TY-5; Small-inducible cytokine A3; SIS-alpha; Macrophage inflammatory protein 1-alpha; L2G25B; Heparin-binding chemotaxis protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546951 375.00
Check your price
  • FOXP3 Monoclonal Antibody, Alexa Fluor® 647 conjugate for Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Alexa Fluor® 647
  • Catalog # MA518160
  • Size 100µg
Applications
Flow Cytometry (6ug/ml)
Alternate names:
Scurfin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518160 349.00
Check your price
  • Nestin Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511887
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
NES, FLJ21841, Nbla00170
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511887 349.00
Check your price
  • C4BPA Monoclonal Antibody for Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA182815
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (1-3ug/ml)
Alternate names:
Proline-rich protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182815 499.00
Check your price
  • CRP Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110320
  • Size 100ug
Applications
ELISA (Assay Dependent)
Alternate names:
RP11-419N10.4, MGC149895, MGC88244, PTX1, C-reactive protein, pentraxin 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110320 249.00
Check your price
  • Mitofilin Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and IP
  • Host Rabbit
  • Target Species Dog, Cat, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA116918
  • Size 100µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunoprecipitation (Assay-Dependent), Western Blot (1:2000)
Alternate names:
Mitofilin; Mitochondrial inner membrane protein; p87/89; Cell proliferation-inducing gene 4/52 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116918 375.00
Check your price