Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  50  60  70  80  90  102  103  104  105  106  107  108  109  110  111  112  113  114  115  116  117  118  119  120  121  122  123  124  125  126  127  128  129  130  131  140  150  160  170  180  190  200  210  300  400  500  600  700  800  900  1000  1100  1630 
3481 - 3510 of 48881 products displayed
  • Rabbit anti-Human IgE (Heavy chain) Secondary Antibody for IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA516396
  • Size 500µL
Applications
Flow Cytometry (1:10), Immunohistochemistry (Paraffin) (1:500-1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516396 149.00
Check your price
  • S6 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532577
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (1:25)
Alternate names:
Phosphoprotein NP33
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532577 349.00
Check your price
  • Cerebellin 2 Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524324
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (8-25 µg/ml)
Alternate names:
CBLN2, UNQ1892/PRO4338, Cerebellin2, Cerebellin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524324 275.00
Check your price
  • Vimentin Monoclonal Antibody for Western Blot, ICC, Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119319
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (1-10µg/ml), Immunocytochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
VIM, FLJ36605
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119319 225.00
Check your price
  • TLR2 Monoclonal Antibody, PE-Cyanine7 conjugate for Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate PE-Cyanine7
  • Catalog # A18453
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Toll/interleukin-1 receptor-like protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18453 Not Orderable Online
Check your price
  • Glucose-6-Phosphate Dehydrogenase Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # PA185474
  • Size 1mL
Applications
ELISA (1/1000-1/5000), Western Blot (Assay Dependent)
Alternate names:
G6PD1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185474 425.00
Check your price
  • AADAT Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532646
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml)
Alternate names:
Kynurenine--oxoglutarate transaminase II; Kynurenine--oxoglutarate transaminase 2; Kynurenine--oxoglutarate aminotransferase II; Kynurenine aminotransferase II; Alpha-aminoadipate aminotransferase; 2-aminoadipate transaminase; 2-aminoadipate aminotransferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532646 349.00
Check your price
  • CD44 Monoclonal Antibody, APC-Cyanine7 conjugate for Flow
  • Host Rat
  • Target Species Human, Mouse
  • Conjugate APC-Cyanine7
  • Catalog # A18641
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Phagocytic glycoprotein I; Phagocytic glycoprotein 1; Lymphocyte antigen 24; Hyaluronate receptor; Hermes antigen; HUTCH-I; GP90 lymphocyte homing/adhesion receptor; Extracellular matrix receptor III; Heparan sulfate proteoglycan; Epican; CDw44
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18641 114.00
Check your price
  • RPS3 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA527974
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
S3, 40S ribosomal protein S3, IMR-90 ribosomal protein S3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527974 349.00
Check your price
  • Phospho-Calmodulin (Thr79, Ser81) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537503
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
CALM1, CALM2, PHKD, PHKD3, caM, calmodulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537503 309.00
Check your price
  • HLA-A2 Monoclonal Antibody for Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110344
  • Size 100ug
Applications
Flow Cytometry (5-10 ug/ml), Immunoprecipitation (Assay dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110344 225.00
Check your price
  • uNOS Polyclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Rabbit
  • Target Species Arthropod, Bovine, Fruit fly, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA1039
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (1:100), Western Blot (1:1,000)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS2; NOS type II; Macrophage NOS; Inducible NO synthase; Hepatocyte NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1039 375.00
Check your price
  • CD3e Monoclonal Antibody for IHC (F) and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA180783
  • Size 100µg
Applications
Flow Cytometry (1/50-1/100), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
T-cell surface antigen T3/Leu-4 epsilon chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180783 159.00
Check your price
  • alpha Actinin Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Hamster, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517308
  • Size 100µL
Applications
Immunocytochemistry (1:25), Immunofluorescence (1:25), Western Blot (1:1000)
Alternate names:
Non-muscle alpha-actinin-1; F-actin cross-linking protein; Alpha-actinin cytoskeletal isoform
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517308 275.00
Check your price
  • NGAL Monoclonal Antibody for Western Blot, IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB2110202
  • Size 200µl
Applications
ELISA (1:8,000), Immunohistochemistry (Assay dependent), Western Blot (1:50)
Alternate names:
p25; Siderocalin LCN2; Oncogene 24p3; Lipocalin-2; 25 kDa alpha-2-microglobulin-related subunit of MMP-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB2110202 650.00
Check your price
  • PDI Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Yeast
  • Conjugate Unconjugated
  • Catalog # MA110032
  • Size 500µL
Applications
Immunocytochemistry (1:500-1:1000), Immunofluorescence (1:500-1:1000), Immunohistochemistry (1:500-1:1000), Western Blot (1:10,000)
Alternate names:
Thioredoxin-related glycoprotein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110032 349.00
Check your price
  • ROBO2 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547461
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
SAX3, Drosophila) homolog 2, roundabout, axon guidance receptor, homolog 2 (Drosophila)
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547461 349.00
Check your price
  • CD3e Monoclonal Antibody for Western Blot, IF, IHC (F), Flow and IP
  • Host Hamster
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA184019
  • Size 250µg
Applications
Flow Cytometry (1:25-1:200), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
T-cell surface antigen T3/Leu-4 epsilon chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA184019 349.00
Check your price
  • HSV Type 2 Polyclonal Antibody, FITC conjugate for IF
  • Host Sheep
  • Target Species Virus
  • Conjugate FITC
  • Catalog # PA17490
  • Size 1mL
Applications
Immunofluorescence (1:10)
Alternate names:
HSV2, HSV-2, Human herpes virus 2, HHV-2, Herpes Simplex II, HSV Type 2, Herpes Simplex 2, HSV Type II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA17490 309.00
Check your price
  • MX2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546707
  • Size 100µl
Applications
Western Blot (1 µg/ml )
Alternate names:
p78-related protein; Myxovirus resistance protein 2; Interferon-regulated resistance GTP-binding protein MxB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546707 309.00
Check your price
  • Firefly luciferase Monoclonal Antibody for Western Blot, IHC (F), Flow and ELISA
  • Host Mouse
  • Target Species Luciferase
  • Conjugate Unconjugated
  • Catalog # MA180225
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (10 ug/ml), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (1/100)
Alternate names:
Luciferin 4-monooxygenase, Firefly Luciferase, Green Luciferase, Japanese firefly, Genji firefly, Photinus pyralis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180225 449.00
Check your price
  • CD19 Monoclonal Antibody, TRI-COLOR® conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate TRI-COLOR®
  • Catalog # MHCD1906
  • Size 0.5ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
T-cell surface antigen Leu-12; Differentiation antigen CD19; B-lymphocyte surface antigen B4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD1906 305.00
Check your price
  • GFAP Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA135377
  • Size 1mL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Immunohistochemistry (Paraffin, Frozen) (1:10-1:50), Western Blot (Assay dependent)
Alternate names:
GFAP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135377 309.00
Check your price
  • CD8 alpha Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA517394
  • Size 500ug
Applications
Flow Cytometry (0.1 ug/10^6 cells), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
OX-8 membrane antigen; CD8 antigen 32 kDa chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517394 309.00
Check your price
  • QSOX1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538009
  • Size 100µg
Applications
Western Blot (1-2 µg/ml)
Alternate names:
Skin sulfhydryl oxidase; Quiescin Q6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538009 309.00
Check your price
  • Nitrated-alpha Synuclein (Tyr125, Tyr133) Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA516142
  • Size 100µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (1:200-1:500), Western Blot (1:500-1:1500)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516142 349.00
Check your price
  • CD25 Monoclonal Antibody, Biotin conjugate for Western Blot and Flow
  • Host Rat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA517679
  • Size 100ug
Applications
Flow Cytometry (0.5 ug/10^6 cells), Western Blot (Assay Dependent)
Alternate names:
p55; TAC antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517679 225.00
Check your price
  • Cyclin D1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA512255
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
PRAD1 oncogene; BCL-1 oncogene; B-cell lymphoma 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512255 349.00
Check your price
  • Phospho-PPAR alpha (Ser12) Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1820
  • Size 100µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:100-1:1000)
Alternate names:
Nuclear receptor subfamily 1 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1820 375.00
Check your price
  • Influenza A H6N1 NP Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA141060
  • Size 100µg
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Influenza H6N1, H6N1 NP, H6N1 Nuclear Protein,
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141060 325.00
Check your price