Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  20  30  40  50  60  70  80  90  100  107  108  109  110  111  112  113  114  115  116  117  118  119  120  121  122  123  124  125  126  127  128  129  130  131  132  133  134  135  136  140  150  160  170  180  190  200  210  220  300  400  500  600  700  800  900  1000  1100  1630 
3631 - 3660 of 48881 products displayed
  • AKT2 Monoclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 393900
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (1:50-1:500), Western Blot (Assay Dependent)
Alternate names:
RAC-PK-beta; Protein kinase B beta; Protein kinase Akt-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
393900 302.00
Check your price
  • Hepatitis B Virus Core Antigen Monoclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17609
  • Size 100µg
Applications
ELISA (1:1000), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
HepB, Hepatitis B virus nucleocapsid particles, HBcAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17609 275.00
Check your price
  • S100A9 Polyclonal Antibody for IF, ICC, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA146489
  • Size 100µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:1000)
Alternate names:
S100 calcium-binding protein A9; Migration inhibitory factor-related protein 14; Leukocyte L1 complex heavy chain; Calgranulin-B; Myeloid-related protein 14; Calprotectin L1H subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146489 309.00
Check your price
  • IL-6 Receptor Monoclonal Antibody for Flow, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # AHR0061
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Neutralization (Assay Dependent)
Alternate names:
Membrane glycoprotein 80; IL-6R 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHR0061 305.00
Check your price
  • Goat anti-Rabbit IgG Fc Cross-Adsorbed Secondary Antibody, FITC conjugate for IF and Flow
  • Host Goat
  • Target Species Rabbit
  • Conjugate FITC
  • Catalog # A16125
  • Size 2mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16125 115.00
Check your price
  • 5-Methylcytidine Monoclonal Antibody for IF, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA180331
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180331 550.00
Check your price
  • HIV1 p17 Monoclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA518032
  • Size 100µL
Applications
Flow Cytometry (Assay-dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (Assay-dependent)
Alternate names:
Human immunodeficiency virus
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518032 399.00
Check your price
  • IKK gamma Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA528606
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
NF-kappa-B essential modifier; Inhibitor of nuclear factor kappa-B kinase subunit gamma; IkB kinase-associated protein 1; FIP-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528606 349.00
Check your price
  • SEMA5A Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Sheep
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA547791
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Semaphorin-F
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547791 349.00
Check your price
  • CA19-9 Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513275
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (2-4 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513275 249.00
Check your price
  • Phospho-FOXA2 (Thr156) Oligoclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710680
  • Size 100µg
Applications
Immunocytochemistry (3-5 µg/ml), Immunofluorescence (3-5 µg/ml), Western Blot (2-3 µg/ml)
Alternate names:
Transcription factor 3B; Forkhead box protein A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
710680 304.00
Check your price
  • ADAM9 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517080
  • Size 100µL
Applications
Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
Myeloma cell metalloproteinase; Metalloprotease/disintegrin/cysteine-rich protein 9; Meltrin-gamma; Cellular disintegrin-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517080 275.00
Check your price
  • TIGIT Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA545848
  • Size 100µl
Applications
Western Blot (1 µg/ml)
Alternate names:
V-set and transmembrane domain-containing protein 3; V-set and immunoglobulin domain-containing protein 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA545848 309.00
Check your price
  • MVP Monoclonal Antibody for ICC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135743
  • Size 100µg
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:20), Immunohistochemistry (Paraffin, Frozen) (1:20)
Alternate names:
Lung resistance-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135743 600.00
Check your price
  • c-Kit Monoclonal Antibody for IF, ICC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512944
  • Size 500µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent)
Alternate names:
v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog; p145 c-kit; Tyrosine-protein kinase Kit; Proto-oncogene c-Kit; Piebald trait protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512944 309.00
Check your price
  • DMPK Polyclonal Antibody for Western Blot, IF and ELISA
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 349900
  • Size 50µg
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Myotonic dystrophy protein kinase; DMPK; DM-kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
349900 304.00
Check your price
  • NDUFA9 Monoclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 459100
  • Size 100µg
Applications
Western Blot (1.0 ug/ml)
Alternate names:
NADH-ubiquinone oxidoreductase 39 kDa subunit; Complex I-39kD; Sperm flagella protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
459100 358.00
Check your price
  • Phospho-ATM (Ser1981) Monoclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA12020
  • Size 200µg
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunoprecipitation (Assay dependent), Western Blot (1:500-1:1000)
Alternate names:
Ataxia telangiectasia mutated; Ataxia telangiectasia mutated homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA12020 299.00
Check your price
  • HA Tag Monoclonal Antibody, DyLight 800 4X PEG conjugate for Western Blot
  • Host Mouse
  • Target Species Tag
  • Conjugate DyLight 800 4X PEG
  • Catalog # 26183D800
  • Size 50µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
26183D800 159.00
Check your price
  • RPAIN Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520327
  • Size 100ug
Applications
Western Blot (0.5-1 ug/ml)
Alternate names:
HRIP, RIP; RAP interaction protein; RPA-interacting protein; nuclear transporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520327 349.00
Check your price
  • GPR19 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1151
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:500-1:5000)
Alternate names:
GPR-NGA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1151 309.00
Check your price
  • MX2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536985
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Myxovirus resistance protein 2; p78-related protein; Interferon-regulated resistance GTP-binding protein MxB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536985 309.00
Check your price
  • B-Myb Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA546845
  • Size 100µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (0.2-1 µg/ml)
Alternate names:
Myb-like protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546845 309.00
Check your price
  • CD163 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # A18360
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Hemoglobin scavenger receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18360 103.00
Check your price
  • CYP39A1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538909
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
Oxysterol 7-alpha-hydroxylase; Cytochrome P450 39A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538909 309.00
Check your price
  • E-selectin Monoclonal Antibody, DyLight® 650 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate DyLight® 650
  • Catalog # MA516071
  • Size 100Tests
Applications
Flow Cytometry (1:10-1:1000)
Alternate names:
Leukocyte-endothelial cell adhesion molecule 2; Endothelial leukocyte adhesion molecule 1; CD62 antigen-like family member E
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516071 375.00
Check your price
  • Complement Factor H Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524425
  • Size 400µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
H factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524425 349.00
Check your price
  • CD45.2 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate PE
  • Catalog # A16327
  • Size 50µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
B220, CD45, CD45R, GP180, L-CA, LCA, LY5, T200, CD45 antigen, T200 glycoprotein, T200 leukocyte common antigen, protein tyrosine phosphatase, receptor type, c polypeptide, receptor-type tyrosine-protein phosphatase C, PTPRC
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16327 129.00
Check your price
  • Cav1.2 Polyclonal Antibody for Western Blot, IP and Neu
  • Host Rabbit
  • Target Species Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA523013
  • Size 100µL
Applications
Blocking Assay (50-200 molar excess), Immunoprecipitation (1:10-1:500), Neutralization (50-200 molar excess), Western Blot (1:500-1:1000)
Alternate names:
Voltage-gated calcium channel subunit alpha Cav1.2; Smooth muscle calcium channel blocker receptor; Calcium channel, L type, alpha-1 polypeptide, isoform 1, cardiac muscle; Rat brain class C; Mouse brain class C; MELC-CC
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523013 349.00
Check your price
  • RSV Fusion Protein Monoclonal Antibody for IF and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17286
  • Size 100µg
Applications
ELISA (1:20), Immunofluorescence (1:10)
Alternate names:
human respiratory syncytial virus, RSV, RSV Fusion protein, Rsvgp08
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17286 309.00
Check your price