Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  20  30  40  50  60  70  80  90  100  111  112  113  114  115  116  117  118  119  120  121  122  123  124  125  126  127  128  129  130  131  132  133  134  135  136  137  138  139  140  150  160  170  180  190  200  210  220  300  400  500  600  700  800  900  1000  1100  1630 
3751 - 3780 of 48881 products displayed
  • Nuclear Matrix Protein p84 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), IP and ChIP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527816
  • Size 100µL
Applications
ChIP assay (Assay Dependent), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (Assay-Dependent), Immunoprecipitation (1:100-1:500), Western Blot (1:500-1:3000)
Alternate names:
Nuclear matrix protein p84; hTREX84
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527816 349.00
Check your price
  • CD64 Monoclonal Antibody, TRI-COLOR® conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate TRI-COLOR®
  • Catalog # CD6406
  • Size 0.5ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Fc-gamma RIA; Fc-gamma RI
Catalog # Size Price (AUD) Your Price (AUD) Qty
CD6406 321.00
Check your price
  • RAB9A Monoclonal Antibody for Western Blot, IF, ICC, IHC, IP and IM
  • Host Mouse
  • Target Species Bovine, Dog, Hamster, Human, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA3067
  • Size 100µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (1:20), Immunomicroscopy (Assay Dependent), Immunoprecipitation (Assay dependent), Western Blot (1:1000-1:5000)
Alternate names:
Ras-related protein Rab-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3067 375.00
Check your price
  • CD19 Monoclonal Antibody, APC-Cyanine5.5 conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate APC-Cyanine5.5
  • Catalog # RM7719
  • Size 500µL
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Differentiation antigen CD19
Catalog # Size Price (AUD) Your Price (AUD) Qty
RM7719 338.00
Check your price
  • KGF Monoclonal Antibody for IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Dog
  • Conjugate Unconjugated
  • Catalog # MA524009
  • Size 100µg
Applications
Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Immunohistochemistry (Paraffin) (8-25 µg/ml)
Alternate names:
Keratinocyte growth factor; Heparin-binding growth factor 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524009 275.00
Check your price
  • CD69 Monoclonal Antibody, PerCP conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PerCP
  • Catalog # MA110277
  • Size 100Tests
Applications
Flow Cytometry (10 ul/10^6 cells)
Alternate names:
MLR-3; Leukocyte surface antigen Leu-23; GP32/28; Early T-cell activation antigen p60; EA1; C-type lectin domain family 2 member C; BL-AC/P26; Activation inducer molecule
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110277 275.00
Check your price
  • CD3 Monoclonal Antibody for IHC (F), IHC (P), Flow and IP
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA180469
  • Size 200µg
Applications
Flow Cytometry (1:10-1:25), Immunohistochemistry (Paraffin, Frozen) (1:10-1:25), Immunoprecipitation (Assay Dependent)
Alternate names:
T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180469 309.00
Check your price
  • Dectin 1 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA516695
  • Size 100TESTS
Applications
Flow Cytometry (Neat)
Alternate names:
Dendritic cell-associated C-type lectin 1; C-type lectin superfamily member 12; Beta-glucan receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516695 349.00
Check your price
  • SLC22A12 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA542510
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Urate anion exchanger 1; Renal-specific transporter; Organic anion transporter 4-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542510 309.00
Check your price
  • Fibroblast Growth Factor basic for Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP10915
  • Size 50µg
Applications
Control (Assay Dependent)
Alternate names:
FGF-b
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP10915 345.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IF, IHC (P), IP and ELISA
  • Host Goat
  • Target Species Rabbit
  • Conjugate HRP
  • Catalog # 656120
  • Size 1mL
Applications
ELISA (1:3,000-1:4,000), Immunofluorescence (1:3,000-1:4,000), Immunohistochemistry (Paraffin) (1:3,000-1:4,000), Immunoprecipitation (1:3,000-1:4,000), Western Blot (1:3,000-1:4,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
656120 149.00
Check your price
  • E. coli J5 LPS Polyclonal Antibody for ELISA
  • Host Sheep
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # PA185648
  • Size 1mL
Applications
ELISA (Assay Dependent)
Alternate names:
Escherichia coli
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185648 349.00
Check your price
  • Rabbit anti-Rat IgG Secondary Antibody, Texas Red® conjugate for IHC (F), IHC (P) and Flow
  • Host Rabbit
  • Target Species Rat
  • Conjugate Texas Red®
  • Catalog # PA128571
  • Size 1mg
Applications
Flow Cytometry (Assay-dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay-dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128571 199.00
Check your price
  • NPR1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529049
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Guanylate cyclase A; Atrial natriuretic peptide receptor type A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529049 349.00
Check your price
  • GNA12 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529928
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
NNX3, RMP, gep, G-protein subunit alpha-12, WUGSC:H_GS165O14.2, g alpha-12, guanine nucleotide-binding protein subunit alpha-12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529928 349.00
Check your price
  • PYY Polyclonal Antibody, Biotin conjugate for ELISA
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # PA19590
  • Size 100µL
Applications
ELISA (1:2000-1:10000)
Alternate names:
Peptide tyrosine tyrosine; PYY-I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19590 375.00
Check your price
  • Staphylococcus Enterotoxin pan Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA183468
  • Size 200µg
Applications
ELISA (1/5,000-1/20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183468 375.00
Check your price
  • CD200R Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate PE
  • Catalog # MA516895
  • Size 100TESTS
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
OX102 antigen; Cell surface glycoprotein OX2 receptor 1; CD200 cell surface glycoprotein receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516895 399.00
Check your price
  • RNF12 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529530
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Renal carcinoma antigen NY-REN-43; RING finger protein 12; RING finger LIM domain-binding protein; LIM domain-interacting RING finger protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529530 349.00
Check your price
  • PAK1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA529256
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
p68-PAK; p21-activated kinase 1; Protein kinase MUK2; Alpha-PAK; p65-PAK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529256 349.00
Check your price
  • Rabies Virus Monoclonal Antibody for IF, ICC and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17079
  • Size 1mL
Applications
ELISA (1:20-1:200), Immunocytochemistry (1:10-50), Immunofluorescence (1:10-50)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17079 349.00
Check your price
  • Cytomegalovirus gB Late Antigen Monoclonal Antibody for Western Blot, IF and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17292
  • Size 100µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50), Western Blot (1:10-1:50)
Alternate names:
CMV, HHV5gp061
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17292 309.00
Check your price
  • Phospho-TEK (Tyr1108) Polyclonal Antibody for Western Blot and IF
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538339
  • Size 100µg
Applications
Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
p140 TEK; Tyrosine-protein kinase receptor TIE-2; Tyrosine-protein kinase receptor TEK; Tyrosine kinase with Ig and EGF homology domains-2; Tunica interna endothelial cell kinase; Endothelial tyrosine kinase; STK1; HYK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538339 309.00
Check your price
  • CD73 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527336
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Ecto-5'-nucleotidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527336 349.00
Check your price
  • Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor® 594 conjugate for IF, ICC, IHC and Flow
  • Host Donkey
  • Target Species Mouse
  • Conjugate Alexa Fluor® 594
  • Catalog # A21203
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21203 209.00
Check your price
  • CD22 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527928
  • Size 100µL
Applications
Western Blot (1:1000-1:10,000)
Alternate names:
T-cell surface antigen Leu-14; Sialic acid-binding Ig-like lectin 2; B-lymphocyte cell adhesion molecule
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527928 349.00
Check your price
  • Phospho-p70 S6 Kinase (Ser418) Polyclonal Antibody for DB
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512782
  • Size 400µL
Applications
Dot blot (1:500)
Alternate names:
p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I; 70 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512782 349.00
Check your price
  • COX4 Polyclonal Antibody for Western Blot, IHC (P), Flow and IP
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517511
  • Size 100µL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (1:2400), Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
Cytochrome c oxidase subunit IV isoform 1; Cytochrome c oxidase polypeptide IV
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517511 275.00
Check your price
  • Phospho-SMAD3 (Ser423, Ser425) Monoclonal Antibody for Western Blot, IP and ChIP
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate, Rat, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA514936
  • Size 100µL
Applications
ChIP assay (1:100), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
SMAD family member 3; JV15-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514936 309.00
Check your price
  • MOG Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518351
  • Size 100ug
Applications
Western Blot (0.03-1 ug/ml)
Alternate names:
MOG; MGC26137; myelin oligodendrocyte glycoprotein; DAQB-92E24.2; MOGIG-2; MOG Ig-AluB; MOG alpha-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518351 349.00
Check your price