Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  30  40  50  60  70  80  90  100  110  114  115  116  117  118  119  120  121  122  123  124  125  126  127  128  129  130  131  132  133  134  135  136  137  138  139  140  141  142  143  150  160  170  180  190  200  210  220  300  400  500  600  700  800  900  1000  1100  1605 
3841 - 3870 of 48136 products displayed
  • QSOX2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534151
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml)
Alternate names:
Quiescin Q6-like protein 1; Neuroblastoma-derived sulfhydryl oxidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534151 349.00
Check your price
  • NOX2 Antibody (Monoclonal, CL5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518050
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
p22 phagocyte B-cytochrome; gp91-phox; gp91-1; Superoxide-generating NADPH oxidase heavy chain subunit; Neutrophil cytochrome b 91 kDa polypeptide; NADPH oxidase 2; Heme-binding membrane glycoprotein gp91phox; Cytochrome b(558) subunit beta; CGD91-phox
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518050 399.00
Check your price
  • CD3 Antibody (Monoclonal, 4D10A6)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517043
  • Size 100 µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000)
Alternate names:
T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain; T-cell receptor T3 zeta chain; T-cell surface antigen T3/Leu-4 epsilon chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517043 225.00
Check your price
  • pro-TGF beta Antibody
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA129020
  • Size 100 µg
Applications
Immunohistochemistry (1:200-1:500), Western Blot (1:2000)
Alternate names:
TGF-beta1, TGFbeta1, Tgfb, Tgfb-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129020 309.00
Check your price
  • Bovine Serum Albumin Antibody (Monoclonal, 5G10)
  • Host Mouse
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # HYB2610202
  • Size 200 µL
Applications
ELISA (Assay Dependent)
Alternate names:
BSA
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB2610202 425.00
Check your price
  • Phospho-mTOR (Ser2448) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 441125G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Rapamycin target protein 1; Rapamycin and FKBP12 target 1; Mechanistic target of rapamycin; Mammalian target of rapamycin; FKBP12-rapamycin complex-associated protein; FK506-binding protein 12-rapamycin complex-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
441125G 308.00
Check your price
  • RELM alpha Antibody (Monoclonal, 226033)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523898
  • Size 100 µg
Applications
Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
RELMalpha; Parasite-induced macrophage novel gene 1 protein; Hypoxia-induced mitogenic factor; Cysteine-rich secreted protein FIZZ1; Cysteine-rich secreted protein A12-gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523898 275.00
Check your price
  • Jagged1 Antibody
  • Host Goat
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA128001
  • Size 25µg
Applications
ELISA (0.5-1 µg/ml), Flow Cytometry (20 µg/ml for 10^6 cells), Neutralization (2-10 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
protein jagged-1, JAG1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128001 375.00
Check your price
  • Ly-6C Antibody (Monoclonal, ER-MP20)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA181899
  • Size 100 µg
Applications
Flow Cytometry (1/10-1/50), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
AA682074, AA959465, Ly-6C, Ly6c, Ly6c2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181899 199.00
Check your price
  • SMC1 Antibody (Monoclonal, C2M)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA112604
  • Size 100 µg
Applications
Immunofluorescence (5-10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
Sb1.8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112604 309.00
Check your price
  • ELOVL6 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520520
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Very long chain 3-oxoacyl-CoA synthase 6; Very long chain 3-ketoacyl-CoA synthase 6; Myelin-associated SUR4 protein; Long chain fatty acid elongase; Fatty acyl-CoA elongase; ELOVL fatty acid elongase 6; 3-keto acyl-CoA synthase Elovl6; Long-chain fatty-acyl elongase; Fatty acid elongase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520520 349.00
Check your price
  • C3a Receptor Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529979
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
AZ3B, C3AR, HNFAG09, C3a anaphylatoxin chemotactic receptor, C3a-R, complement component 3 receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529979 Not Orderable Online
Check your price
  • Phospho-GluR1 (Ser831) Antibody
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA184839
  • Size 150 µL
Applications
Western Blot (1:1000)
Alternate names:
Glutamate receptor ionotropic, AMPA 1; GluR-K1; GluR-A; AMPA-selective glutamate receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184839 650.00
Check your price
  • MVP Antibody (Monoclonal, MVP-37)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135591
  • Size 1 mL
Applications
Flow Cytometry (Assay dependent.), Immunohistochemistry (Paraffin, Frozen) (1:20), Western Blot (1:50)
Alternate names:
Lung resistance-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135591 550.00
Check your price
  • CD162 Antibody (Monoclonal, TC2)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110114
  • Size 100 ug
Applications
Flow Cytometry (1.5-2.5 ug/ml)
Alternate names:
Selectin P ligand
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110114 159.00
Check your price
  • SSEA1 Antibody (Monoclonal, MC-480)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 411200
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent)
Alternate names:
stage-specific embryonic antigen 1, SSEA1, MC-480, CD15, SSEA-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
411200 309.00
Check your price
  • SLC27A1 Antibody (Monoclonal, 308420)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524022
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml)
Alternate names:
Solute carrier family 27 member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524022 275.00
Check your price
  • alpha Synuclein Antibody (Monoclonal, 14H2L1)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701085
  • Size 100 µg
Applications
Flow Cytometry (1:100-1:500), Immunocytochemistry (1:500-1:5000), Immunofluorescence (1:500-1:5000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:5000)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
701085 304.00
Check your price
  • GDF11 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511928
  • Size 400 µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Bone morphogenetic protein 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511928 349.00
Check your price
  • E-cadherin Antibody (Monoclonal, DH01 (DCS-266))
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511496
  • Size 500 µL
Applications
Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunoprecipitation (2 µg/mg protein lysate), Western Blot (1-2 µg/ml)
Alternate names:
Uvomorulin; Epithelial cadherin; CAM 120/80
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511496 349.00
Check your price
  • Goat anti-Armenian Hamster IgG, AP
  • Host Goat
  • Target Species Hamster
  • Conjugate AP
  • Catalog # PA129558
  • Size 500 µg
Applications
ELISA (1:2000-1:10,000), Western Blot (1:500-1:2500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129558 275.00
Check your price
  • MiTF Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538294
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Class E basic helix-loop-helix protein 32
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538294 309.00
Check your price
  • ERCC1 Antibody (Monoclonal, 8F1)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA513912
  • Size 500 µL
Applications
Flow Cytometry (0.5-1ug/test), Immunohistochemistry (Paraffin) (1:100-1:500), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
DNA excision repair protein ERCC-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513912 349.00
Check your price
  • Complement C3d Antibody (Monoclonal, 7C10)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HAV0030502
  • Size 200 µl
Applications
ELISA (1:16,000), Western Blot (Assay dependent)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
HAV0030502 425.00
Check your price
  • ESRP2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532143
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
RNA-binding protein 35B; RNA-binding motif protein 35B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532143 349.00
Check your price
  • ULBP2/ULBP5/ULBP6 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547118
  • Size 100 µg
Applications
Blocking Assay (Assay-Dependent), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
UL-16 binding protein 5; UL16-binding protein 2; Retinoic acid early transcript 1H; ALCAN-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547118 375.00
Check your price
  • ALK Antibody (Monoclonal, 5A4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511460
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:40-1:80)
Alternate names:
Anaplastic lymphoma kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511460 475.00
Check your price
  • RASSF9 Antibody (Monoclonal, 2F8)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA520630
  • Size 100 µg
Applications
ELISA (Assay-Dependent)
Alternate names:
Peptidylglycine alpha-amidating monooxygenase COOH-terminal interactor; PAM COOH-terminal interactor protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA520630 309.00
Check your price
  • GPR1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132815
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132815 375.00
Check your price
  • Goat anti-Mouse IgG, IgM (H+L), Biotin
  • Host Goat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # 31807
  • Size 2 mL
Applications
ELISA (1:20,000-1:400,000), Flow Cytometry (1:200-1:1,000), Immunocytochemistry (1:500-1:5,000), Immunofluorescence (1:500-1:5,000), Immunohistochemistry (1:500-1:5,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:20,000-1:400,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31807 173.00
Check your price