Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  30  40  50  60  70  80  90  100  110  122  123  124  125  126  127  128  129  130  131  132  133  134  135  136  137  138  139  140  141  142  143  144  145  146  147  148  149  150  151  160  170  180  190  200  210  220  230  300  400  500  600  700  800  900  1000  1100  1611 
4081 - 4110 of 48310 products displayed
  • pro-TGF beta Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA129020
  • Size 100 µg
Applications
Immunohistochemistry (1:200-1:500), Western Blot (1:2000)
Alternate names:
TGF-beta1, TGFbeta1, Tgfb, Tgfb-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129020 309.00
Check your price
  • CREST Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530500
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
SS18-like protein 1; SYT homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530500 349.00
Check your price
  • Jagged1 Polyclonal Antibody for Western Blot, Flow, ELISA and Neu
  • Host Goat
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA128001
  • Size 25µg
Applications
ELISA (0.5-1 µg/ml), Flow Cytometry (20 µg/ml for 10^6 cells), Neutralization (2-10 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
protein jagged-1, JAG1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128001 Not Orderable Online
Check your price
  • Ly-6C Monoclonal Antibody for IHC (F), Flow and IP
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA181899
  • Size 100 µg
Applications
Flow Cytometry (1/10-1/50), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
AA682074, AA959465, Ly-6C, Ly6c, Ly6c2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181899 199.00
Check your price
  • CD357 Monoclonal Antibody, APC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate APC
  • Catalog # A18630
  • Size 25 µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Glucocorticoid-induced TNFR-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18630 155.00
Check your price
  • ELOVL6 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520520
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Very long chain 3-oxoacyl-CoA synthase 6; Very long chain 3-ketoacyl-CoA synthase 6; Myelin-associated SUR4 protein; Long chain fatty acid elongase; Fatty acyl-CoA elongase; ELOVL fatty acid elongase 6; 3-keto acyl-CoA synthase Elovl6; Long-chain fatty-acyl elongase; Fatty acid elongase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520520 349.00
Check your price
  • AFP Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB0970402
  • Size 200 µl
Applications
ELISA (1:7,500)
Alternate names:
Alpha-fetoglobulin; Alpha-1-fetoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB0970402 550.00
Check your price
  • MVP Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135591
  • Size 1 mL
Applications
Flow Cytometry (Assay dependent.), Immunohistochemistry (Paraffin, Frozen) (1:20), Western Blot (1:50)
Alternate names:
Lung resistance-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135591 550.00
Check your price
  • CD162 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110114
  • Size 100 ug
Applications
Flow Cytometry (1.5-2.5 ug/ml)
Alternate names:
Selectin P ligand
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110114 159.00
Check your price
  • SSEA1 Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 411200
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent)
Alternate names:
stage-specific embryonic antigen 1, SSEA1, MC-480, CD15, SSEA-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
411200 309.00
Check your price
  • PMS2 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529835
  • Size 100 µL
Applications
Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
PMS1 protein homolog 2; DNA mismatch repair protein PMS2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529835 349.00
Check your price
  • TRAF3 Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 700121
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (0.5-1µg/ml)
Alternate names:
LMP1-associated protein 1; CD40-binding protein; CD40 receptor-associated factor 1; CAP-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
700121 309.00
Check your price
  • GDF11 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511928
  • Size 400 µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Bone morphogenetic protein 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511928 349.00
Check your price
  • alpha Synuclein Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701085
  • Size 100 µg
Applications
Flow Cytometry (1:100-1:500), Immunocytochemistry (1:500-1:5000), Immunofluorescence (1:500-1:5000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:5000)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
701085 304.00
Check your price
  • F(ab')2-Goat anti-Human IgA Secondary Antibody, FITC conjugate for IF and Flow
  • Host Goat
  • Target Species Human
  • Conjugate FITC
  • Catalog # A24459
  • Size 500 µg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24459 150.00
Check your price
  • E-cadherin Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511496
  • Size 500 µL
Applications
Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunoprecipitation (2 µg/mg protein lysate), Western Blot (1-2 µg/ml)
Alternate names:
Uvomorulin; Epithelial cadherin; CAM 120/80
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511496 349.00
Check your price
  • LOXL2 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523963
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Lysyl oxidase-related protein WS9-14; Lysyl oxidase-related protein 2; Lysyl oxidase-like protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523963 275.00
Check your price
  • Medroxyprogesterone Acetate Polyclonal Antibody for ELISA
  • Host Sheep
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # PA175249
  • Size 100 µL
Applications
ELISA (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175249 600.00
Check your price
  • MiTF Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538294
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Class E basic helix-loop-helix protein 32
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538294 309.00
Check your price
  • Complement C3d Monoclonal Antibody for Western Blot, IF, IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HAV0030502
  • Size 200 µl
Applications
ELISA (1:16,000), Western Blot (Assay dependent)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
HAV0030502 425.00
Check your price
  • GST Omega 1 140A (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP75564
  • Size 100 µg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75564 473.00
Check your price
  • ESRP2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532143
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
RNA-binding protein 35B; RNA-binding motif protein 35B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532143 349.00
Check your price
  • Pirh2 Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA515596
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
p53-induced RING-H2 protein; Zinc finger protein 363; RING finger protein 199; E3 ubiquitin-protein ligase Pirh2; CH-rich-interacting match with PLAG1; Androgen receptor N-terminal-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515596 349.00
Check your price
  • Collagen IV Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514100
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
COL4A1 NC1 domain,collagen IV, alpha-1 polypeptide, collagen alpha-1(IV) chain, collagen of basement membrane, alpha-1 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514100 309.00
Check your price
  • RASSF9 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA520630
  • Size 100 µg
Applications
ELISA (Assay-Dependent)
Alternate names:
Peptidylglycine alpha-amidating monooxygenase COOH-terminal interactor; PAM COOH-terminal interactor protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA520630 309.00
Check your price
  • GPR1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132815
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132815 375.00
Check your price
  • Goat anti-Mouse IgG, IgM (H+L) Secondary Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # 31807
  • Size 2 mL
Applications
ELISA (1:20,000-1:400,000), Flow Cytometry (1:200-1:1,000), Immunocytochemistry (1:500-1:5,000), Immunofluorescence (1:500-1:5,000), Immunohistochemistry (1:500-1:5,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:20,000-1:400,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31807 173.00
Check your price
  • TLR2 Monoclonal Antibody, PE conjugate for IF, ICC, Flow and IP
  • Host Mouse
  • Target Species Dog, Human
  • Conjugate PE
  • Catalog # MA516204
  • Size 100 µl
Applications
Flow Cytometry (0.5-2 µg/10^6 cells), Immunocytochemistry (1:10-1:2000), Immunofluorescence (1:10-1:2000), Immunoprecipitation (2-5 µg/ml)
Alternate names:
Toll/interleukin-1 receptor-like protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516204 375.00
Check your price
  • TFAP4 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA540719
  • Size 100 µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (1.25 µg/ml)
Alternate names:
Class C basic helix-loop-helix protein 41; Activating enhancer-binding protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540719 309.00
Check your price
  • FHL2 Monoclonal Antibody for Western Blot, IF, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA140200
  • Size 100 µg
Applications
Immunofluorescence (Assay Dependent.), Immunohistochemistry (Frozen) (Assay Dependent.), Immunohistochemistry (Paraffin) (Assay Dependent.), Western Blot (Assay Dependent.)
Alternate names:
Skeletal muscle LIM-protein 3; LIM domain protein DRAL
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140200 750.00
Check your price