Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  30  40  50  60  70  80  90  100  110  123  124  125  126  127  128  129  130  131  132  133  134  135  136  137  138  139  140  141  142  143  144  145  146  147  148  149  150  151  152  160  170  180  190  200  210  220  230  300  400  500  600  700  800  900  1000  1100  1605 
4111 - 4140 of 48136 products displayed
  • Metadherin Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 406400
  • Size 100 µg
Applications
Flow Cytometry (3-5 ug x 10^6 cells), Immunocytochemistry (1-2 ug/ml), Immunofluorescence (1-2 ug/ml), Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (1-2 ug/ml)
Alternate names:
Metastasis adhesion protein; Metadherin; Lysine-rich CEACAM1 co-isolated protein; Astrocyte elevated gene-1 protein; 3D3/LYRIC
Catalog # Size Price (AUD) Your Price (AUD) Qty
406400 303.00
Check your price
  • CYP11B2 Antibody
  • Host Rabbit
  • Target Species Horse, Human
  • Conjugate Unconjugated
  • Catalog # PA542061
  • Size 100 µl
Applications
Western Blot (1:100)
Alternate names:
Steroid 18-hydroxylase; Cytochrome P-450C18; Cytochrome P-450Aldo; CYPXIB2; Aldosterone-synthesizing enzyme; Aldosterone synthase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542061 309.00
Check your price
  • ITGA8 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547572
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
ITGA8, integrin alpha-8, integrin, alpha 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547572 349.00
Check your price
  • Torc1 Antibody (Monoclonal, 1B5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515666
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Transducer of regulated cAMP response element-binding protein 1; Mucoepidermoid carcinoma translocated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515666 349.00
Check your price
  • F(ab')2-Goat anti-Mouse IgG, Alexa Fluor® 488-FluoroNanogold
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 488-FluoroNanogold
  • Catalog # A24920
  • Size 1 mL
Applications
Immunomicroscopy (0.2-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24920 530.00
Check your price
  • CD52 Antibody, PE (Monoclonal, CF1D12)
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MHCD5204
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Human epididymis-specific protein 5; Epididymal secretory protein E5; Cambridge pathology 1 antigen; CDw52
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD5204 305.00
Check your price
  • NANOS2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520536
  • Size 100 ug
Applications
Western Blot (2-4 µg/ml)
Alternate names:
NOS2; NOS-2; nanos homolog 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520536 349.00
Check your price
  • Bovine Serum Albumin Antibody
  • Host Rabbit
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # PA129262
  • Size 1 mL
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
BSA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129262 249.00
Check your price
  • Actin Antibody (Monoclonal, mAbGEa)
  • Host Mouse
  • Target Species Bovine, Fruit fly, Human, Mouse, Sheep, Plant, Rat, Xenopus, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA1744
  • Size 100 µL
Applications
ELISA (Assay dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (1:100), Western Blot (1:1,000)
Alternate names:
Beta-actin; Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin-1; Cytoplasmic beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1744 375.00
Check your price
  • Estrogen Receptor alpha Antibody (Monoclonal, 6F11)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA180216
  • Size 100 µL
Applications
Immunohistochemistry (Frozen) (1:40-1:60), Immunohistochemistry (Paraffin) (1:40-1:80), Immunoprecipitation (10 µl/mg lysate), Western Blot (1:50-1:100)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180216 159.00
Check your price
  • Goat anti-Guinea Pig IgG (H+L), HRP
  • Host Goat
  • Target Species Guinea pig
  • Conjugate HRP
  • Catalog # A18769
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18769 109.00
Check your price
  • PTGIS Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA542564
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Prostaglandin I2 synthase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542564 309.00
Check your price
  • WNK3 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA543188
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Protein kinase with no lysine 3; Protein kinase lysine-deficient 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543188 309.00
Check your price
  • PACSIN2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA515354
  • Size 400 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Syndapin-II; Syndapin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515354 349.00
Check your price
  • PON1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524178
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Serum aryldialkylphosphatase 1; K-45; Aromatic esterase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524178 349.00
Check your price
  • IGF2R Antibody, PE (Monoclonal, MEM-238)
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # A15775
  • Size 100 tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
M6P/IGF2 receptor; Insulin-like growth factor II receptor; Insulin-like growth factor 2 receptor; 300 kDa mannose 6-phosphate receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15775 169.00
Check your price
  • SHARPIN Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA540129
  • Size 100 µl
Applications
Western Blot (1:500-1:3000)
Alternate names:
Shank-interacting protein-like 1; Shank-associated RH domain-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540129 309.00
Check your price
  • CD44 Antibody (Monoclonal, MEM-263)
  • Host Mouse
  • Target Species Dog, Human, Pig
  • Conjugate Unconjugated
  • Catalog # MA119277
  • Size 100 µg
Applications
Flow Cytometry (4 µg/ml), Immunohistochemistry (Paraffin) (10 µg/ml), Immunoprecipitation (Assay Dependent), Western Blot (2 µg/ml)
Alternate names:
Phagocytic glycoprotein I; Phagocytic glycoprotein 1; Hyaluronate receptor; Hermes antigen; Heparan sulfate proteoglycan; HUTCH-I; GP90 lymphocyte homing/adhesion receptor; Extracellular matrix receptor III; Epican; CDw44
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119277 225.00
Check your price
  • GFP Tag Antibody (Monoclonal)
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # G10362
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Green Fluorescence Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
G10362 291.00
Check your price
  • Diazepam Binding Inhibitor Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA540659
  • Size 100 µl
Applications
Immunohistochemistry (5 µg/ml), Western Blot (0.2-1 µg/ml)
Alternate names:
Endozepine; Diazepam-binding inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540659 309.00
Check your price
  • CD43 Antibody, FITC (Monoclonal, MEM-59)
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA119567
  • Size 100 tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
CD43, GALGP, GPL115, LSN, galactoglycoprotein, leukocyte sialoglycoprotein, leukosialin, sialophorin gpL115, leukosialin, CD43
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119567 225.00
Check your price
  • CD44 Antibody, Biotin (Monoclonal, 156-3C11)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Biotin
  • Catalog # MA513887
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Phagocytic glycoprotein I; Phagocytic glycoprotein 1; Hyaluronate receptor; Hermes antigen; Heparan sulfate proteoglycan; HUTCH-I; GP90 lymphocyte homing/adhesion receptor; Extracellular matrix receptor III; Epican; CDw44
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513887 375.00
Check your price
  • CEACAM6 Antibody (Monoclonal, 439424)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524164
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Normal cross-reacting antigen; Non-specific crossreacting antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524164 275.00
Check your price
  • Lysozyme Antibody (Monoclonal, BGN/06/964)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA183315
  • Size 200 µg
Applications
ELISA (Assay Dependent)
Alternate names:
1,4-beta-N-acetylmuramidase C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183315 325.00
Check your price
  • CD7 Antibody, TRI-COLOR® (Monoclonal, CD7-6B7)
  • Host Mouse
  • Target Species Human
  • Conjugate TRI-COLOR®
  • Catalog # MHCD0706
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
TP41; T-cell surface antigen Leu-9; T-cell leukemia antigen; GP40
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD0706 303.00
Check your price
  • Follicle Stimulating Hormone Antibody
  • Host Rabbit
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # PA185444
  • Size 50 µL
Applications
Immunohistochemistry (Frozen) (1/200-1/500), Western Blot (1/300-1/400)
Alternate names:
Follitropin beta chain; Follicle-stimulating hormone beta subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185444 600.00
Check your price
  • Acinus Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519938
  • Size 100 ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (1 µg/ml)
Alternate names:
ACINUS, ACN, fSAP152; apoptotic chromatin condensation inducer in the nucleus; functional spliceosome-associated protein 152
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519938 349.00
Check your price
  • MRP2 Antibody (Monoclonal, M2II-12)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135677
  • Size 1 mL
Applications
Immunocytochemistry (1:20), Immunohistochemistry (Frozen) (1:20), Western Blot (1:20-1:50)
Alternate names:
Multidrug resistance-associated protein 2; Canalicular multidrug resistance protein; ATP-binding cassette sub-family C member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135677 550.00
Check your price
  • NTE Antibody
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA535939
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (Assay-Dependent)
Alternate names:
Patatin-like phospholipase domain-containing protein 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535939 309.00
Check your price
  • CD10 Antibody (Monoclonal, 2A1H5E1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515231
  • Size 100 ug
Applications
Immunohistochemistry (1/200-1/1000)
Alternate names:
Skin fibroblast elastase; Neutral endopeptidase 24.11; Enkephalinase; Common acute lymphocytic leukemia antigen; Atriopeptidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515231 349.00
Check your price