Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  40  50  60  70  80  90  100  110  120  131  132  133  134  135  136  137  138  139  140  141  142  143  144  145  146  147  148  149  150  151  152  153  154  155  156  157  158  159  160  170  180  190  200  210  220  230  240  300  400  500  600  700  800  900  1000  1100  1630 
4351 - 4380 of 48882 products displayed
  • FOXP3 Polyclonal Antibody for Western Blot, ICC, IHC (P), ChIP and pep-ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA19044
  • Size 100ug
Applications
ChIP assay (Assay dependent), Immunocytochemistry (2-4 ug/ml), Immunohistochemistry (Paraffin) (0.3-1 µg/ml), Western Blot (Assay dependent)
Alternate names:
Scurfin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19044 349.00
Check your price
  • Mouse Granulocyte-Macrophage Colony-Stimulating Factor (GM-CSF) for Western Blot, IP, ELISA and Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RMGMCSF20
  • Size 20ug
Applications
Control (Assay Dependent), ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Colony-stimulating factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
RMGMCSF20 298.00
Check your price
  • SPT3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Goat, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA540315
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (4-8 µg/ml), Western Blot (0.4 µg/ml)
Alternate names:
SPT3-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540315 309.00
Check your price
  • POTEH Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524910
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Prostate, ovary, testis-expressed protein on chromosome 22; ANKRD26-like family C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524910 349.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate AP
  • Catalog # G21060
  • Size 1mg
Applications
ELISA (1:500 to 1:2,000), Immunohistochemistry (1:500 to 1:2,000), Western Blot (1:500 to 1:2,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
G21060 160.00
Check your price
  • Phospho-RSK1 (Thr359, Ser363) Polyclonal Antibody for Western Blot, IP and ChIP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517901
  • Size 100µL
Applications
ChIP assay (1-3 ul), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Ribosomal S6 kinase 1; MAP kinase-activated protein kinase 1a; 90 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517901 309.00
Check your price
  • AKT2 Monoclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 393900
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (1:50-1:500), Western Blot (Assay Dependent)
Alternate names:
RAC-PK-beta; Protein kinase B beta; Protein kinase Akt-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
393900 302.00
Check your price
  • Hepatitis B Virus Core Antigen Monoclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17609
  • Size 100µg
Applications
ELISA (1:1000), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
HepB, Hepatitis B virus nucleocapsid particles, HBcAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17609 275.00
Check your price
  • Phospho-c-Met (Tyr1230, Tyr1234, Tyr1235) Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA128463
  • Size 50µL
Applications
ELISA (Assay-dependent), Western Blot (1:1000)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128463 349.00
Check your price
  • CD205 Monoclonal Antibody, FITC conjugate for IHC (F), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA135934
  • Size 100µg
Applications
Flow Cytometry (1:25-1:50), Immunohistochemistry (Frozen) (Assay dependent.), Immunoprecipitation (Assay dependent.)
Alternate names:
RP23-10L17.4, CD205, DEC-205, ly-75, CLEC13B, GP200-MR6, DCL-1 fusion, LY75-CD302 fusion protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135934 700.00
Check your price
  • HIF1A Polyclonal Antibody for IF, ICC, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA116627
  • Size 100µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:1000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116627 375.00
Check your price
  • HLA-G Monoclonal Antibody, Alexa Fluor® 488 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Alexa Fluor® 488
  • Catalog # MA518164
  • Size 100µg
Applications
Flow Cytometry (15ug/ml)
Alternate names:
MHC class I antigen G; HLA G antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518164 375.00
Check your price
  • Caspase 1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532292
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
p45; Interleukin-1 beta-converting enzyme; Interleukin-1 beta convertase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532292 275.00
Check your price
  • CD56 Monoclonal Antibody, PE-Texas Red® conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE-Texas Red®
  • Catalog # MHCD5617
  • Size 0.5ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD5617 352.00
Check your price
  • CENPH Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA522523
  • Size 100µg
Applications
ELISA (Assay-Dependent), Western Blot (1-5 µg/ml)
Alternate names:
Interphase centromere complex protein 35
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA522523 309.00
Check your price
  • SLC22A3 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537889
  • Size 100µg
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Organic cation transporter 3; Extraneuronal monoamine transporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537889 309.00
Check your price
  • 5-Methylcytidine Monoclonal Antibody for IF, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA180331
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180331 550.00
Check your price
  • HIV1 p17 Monoclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA518032
  • Size 100µL
Applications
Flow Cytometry (Assay-dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (Assay-dependent)
Alternate names:
Human immunodeficiency virus
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518032 399.00
Check your price
  • IKK gamma Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA528606
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
NF-kappa-B essential modifier; Inhibitor of nuclear factor kappa-B kinase subunit gamma; IkB kinase-associated protein 1; FIP-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528606 349.00
Check your price
  • STKLD1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534747
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Sugen kinase 071; Serine/threonine kinase-like domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534747 349.00
Check your price
  • HSD17B4 Monoclonal Antibody for ICC, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A21975
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (4 ug/ml), Immunoprecipitation (Assay Dependent)
Alternate names:
Short chain dehydrogenase/reductase family 8C member 1; Multifunctional protein 2; D-bifunctional protein; 17-beta-hydroxysteroid dehydrogenase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21975 Not Orderable Online
Check your price
  • SEMA5A Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Sheep
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA547791
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Semaphorin-F
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547791 349.00
Check your price
  • Cytokeratin 18 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529609
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:5000-1:20,000)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529609 349.00
Check your price
  • Poliovirus Type 1 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # HYB2951502
  • Size 200µL
Applications
ELISA (Assay Dependent)
Alternate names:
PV-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB2951502 550.00
Check your price
  • PSA Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532565
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Semenogelase; P-30 antigen; Kallikrein-3; Gamma-seminoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532565 159.00
Check your price
  • CA19-9 Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513275
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (2-4 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513275 249.00
Check your price
  • Phospho-FOXA2 (Thr156) Oligoclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710680
  • Size 100µg
Applications
Immunocytochemistry (3-5 µg/ml), Immunofluorescence (3-5 µg/ml), Western Blot (2-3 µg/ml)
Alternate names:
Transcription factor 3B; Forkhead box protein A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
710680 304.00
Check your price
  • Rabbit anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, TRITC conjugate for IF
  • Host Rabbit
  • Target Species Mouse
  • Conjugate TRITC
  • Catalog # A16171
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16171 152.00
Check your price
  • ABCA13 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA542406
  • Size 100µl
Applications
Western Blot (1 µg/ml )
Alternate names:
ATP-binding cassette, sub-family A (ABC1) member 13
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542406 309.00
Check your price
  • GABBR1 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517075
  • Size 100µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
GABA-B receptor 1; GABA-B-R1; GABA-BR1; GABAB, subunit 1c; OTTHUMP00000109099; OTTHUMP00000214725; gamma-aminobutyric acid type B receptor subunit 1; seven transmembrane helix receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517075 275.00
Check your price