Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  60  70  80  90  100  110  120  130  140  149  150  151  152  153  154  155  156  157  158  159  160  161  162  163  164  165  166  167  168  169  170  171  172  173  174  175  176  177  178  180  190  200  210  220  230  240  250  260  300  400  500  600  700  800  900  1000  1100  1605 
4891 - 4920 of 48136 products displayed
  • GSTA1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529811
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:10,000)
Alternate names:
Glutathione S-transferase Ya1; Glutathione S-transferase Ya; GST class-alpha member 1; GTH1; GSTA1-1; GST-epsilon; GST HA subunit 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529811 349.00
Check your price
  • CD43 Antibody, FITC (Monoclonal, MEM-59)
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA119567
  • Size 100 tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
CD43, GALGP, GPL115, LSN, galactoglycoprotein, leukocyte sialoglycoprotein, leukosialin, sialophorin gpL115, leukosialin, CD43
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119567 225.00
Check your price
  • Phospho-Tau (Ser262) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44750G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (1:500-1:2000)
Alternate names:
Paired helical filament-tau; Neurofibrillary tangle protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
44750G 304.00
Check your price
  • CEACAM6 Antibody (Monoclonal, 439424)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524164
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Normal cross-reacting antigen; Non-specific crossreacting antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524164 275.00
Check your price
  • CD4/CD8 Antibody, FITC, PE (Monoclonal, YKIX302.9/YCATE55.9)
  • Host Rat
  • Target Species Dog
  • Conjugate FITC , PE
  • Catalog # MA516990
  • Size 100 TESTS
Applications
Flow Cytometry (Neat)
Alternate names:
T-cell surface antigen T4/Leu-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516990 499.00
Check your price
  • Coronin 6 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA524759
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Coronin-like protein E
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524759 349.00
Check your price
  • Transferrin Receptor Antibody (Monoclonal, 236-15375)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A11130
  • Size 50 µg
Applications
Immunocytochemistry (1.0 ug/ml), Immunofluorescence (Assay Dependent)
Alternate names:
p90; T9
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11130 244.00
Check your price
  • Connexin 36 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 516300
  • Size 100 µg
Applications
ELISA (0.1-1.0 ug/ml), Immunoprecipitation (2-3 ug), Western Blot (1.0 ug/ml)
Alternate names:
Gap junction alpha-9 protein; Connexin-36
Catalog # Size Price (AUD) Your Price (AUD) Qty
516300 305.00
Check your price
  • ETV2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA542249
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Ets-related protein 71
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542249 309.00
Check your price
  • CD7 Antibody, TRI-COLOR® (Monoclonal, CD7-6B7)
  • Host Mouse
  • Target Species Human
  • Conjugate TRI-COLOR®
  • Catalog # MHCD0706
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
TP41; T-cell surface antigen Leu-9; T-cell leukemia antigen; GP40
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD0706 303.00
Check your price
  • Follicle Stimulating Hormone Antibody
  • Host Rabbit
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # PA185444
  • Size 50 µL
Applications
Immunohistochemistry (Frozen) (1/200-1/500), Western Blot (1/300-1/400)
Alternate names:
Follitropin beta chain; Follicle-stimulating hormone beta subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185444 600.00
Check your price
  • Acinus Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519938
  • Size 100 ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (1 µg/ml)
Alternate names:
ACINUS, ACN, fSAP152; apoptotic chromatin condensation inducer in the nucleus; functional spliceosome-associated protein 152
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519938 349.00
Check your price
  • Caspase 6 Antibody (Monoclonal, 3F52)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191593
  • Size 250 µL
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunoprecipitation (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
Apoptotic protease Mch-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191593 375.00
Check your price
  • MRP2 Antibody (Monoclonal, M2II-12)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135677
  • Size 1 mL
Applications
Immunocytochemistry (1:20), Immunohistochemistry (Frozen) (1:20), Western Blot (1:20-1:50)
Alternate names:
Multidrug resistance-associated protein 2; Canalicular multidrug resistance protein; ATP-binding cassette sub-family C member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135677 550.00
Check your price
  • DLX2 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Guinea pig, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA540505
  • Size 100 µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (2.5 µg/ml)
Alternate names:
Homeobox protein TES-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540505 309.00
Check your price
  • CD10 Antibody (Monoclonal, 2A1H5E1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515231
  • Size 100 ug
Applications
Immunohistochemistry (1/200-1/1000)
Alternate names:
Skin fibroblast elastase; Neutral endopeptidase 24.11; Enkephalinase; Common acute lymphocytic leukemia antigen; Atriopeptidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515231 349.00
Check your price
  • PAX5 Antibody (Monoclonal, SP34)
  • Host Rabbit
  • Target Species Dog, Human
  • Conjugate Unconjugated
  • Catalog # MA139420
  • Size 1 mL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (1:10-1:100)
Alternate names:
B-cell-specific transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA139420 700.00
Check your price
  • UBD Antibody
  • Host Rabbit
  • Target Species Bovine, Horse, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA542827
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Ubiquitin-like protein FAT10; Diubiquitin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542827 309.00
Check your price
  • IL-17 A/F Antibody (Monoclonal, 50104)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523748
  • Size 500 µg
Applications
Neutralization (0.05-0.15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Cytotoxic T-lymphocyte-associated antigen 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523748 349.00
Check your price
  • OASL Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534873
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
p59 OASL; Thyroid receptor-interacting protein 14; 59 kDa 2'-5'-oligoadenylate synthase-like protein; 2'-5'-OAS-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534873 349.00
Check your price
  • CD2 Antibody, Qdot® 605 (Monoclonal, S5.5)
  • Host Mouse
  • Target Species Human
  • Conjugate Qdot® 605
  • Catalog # Q10172
  • Size 100 µL
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
T-cell surface antigen T11/Leu-5; Rosette receptor; LFA-3 receptor; LFA-2; Erythrocyte receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
Q10172 518.00
Check your price
  • MAGI3 Antibody
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112324
  • Size 250 µg
Applications
Western Blot (1- 10 µg/ml)
Alternate names:
Membrane-associated guanylate kinase inverted 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112324 309.00
Check your price
  • PLCG1 Antibody (Monoclonal, 1F1)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFMA0049
  • Size 100 µL
Applications
Immunoprecipitation (1 µg), Western Blot (0.1 µg/ml)
Alternate names:
Phospholipase C-gamma-1; Phosphoinositide phospholipase C-gamma-1; Phospholipase C-II; PLC-148
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0049 309.00
Check your price
  • CD11a Antibody (Monoclonal, 87-6F9)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135057
  • Size 100 µg
Applications
Flow Cytometry (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
Leukocyte function-associated molecule 1 alpha chain; Leukocyte adhesion glycoprotein LFA-1 alpha chain; CD11 antigen-like family member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135057 375.00
Check your price
  • Dynactin 1 Antibody (Monoclonal, 6B11)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518308
  • Size 100 µl
Applications
Immunoprecipitation (1:50-1:200), Western Blot (1:1000)
Alternate names:
p150-glued; p135; DAP-150; 150 kDa dynein-associated polypeptide
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518308 399.00
Check your price
  • FAK Antibody (Monoclonal, 34Q36)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # AHO1272
  • Size 100 µg
Applications
Flow Cytometry (1-3 ug x 10^6 cells), Immunocytochemistry (1-2 ug/ml), Immunofluorescence (1-2 ug/ml), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (1-2 ug/ml)
Alternate names:
pp125FAK; p125FAK; Protein-tyrosine kinase 2; Focal adhesion kinase-related nonkinase; Protein phosphatase 1 regulatory subunit 71
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHO1272 304.00
Check your price
  • URO8 Antibody (Monoclonal, F31)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA190193
  • Size 500 µL
Applications
Immunocytochemistry (1:40), Immunofluorescence (1:40), Immunohistochemistry (Frozen) (1:40)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190193 375.00
Check your price
  • RAC2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529681
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
p21-Rac2; Small G protein; GX
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529681 349.00
Check your price
  • TROVE2 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA541791
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (4-8 µg/ml), Western Blot (2.5 µg/ml)
Alternate names:
TROVE domain family member 2; Sjoegren syndrome type A antigen; Sjoegren syndrome antigen A2; Ro 60 kDa autoantigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541791 309.00
Check your price
  • Glu-Glu Tag Antibody
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # PA517335
  • Size 100 µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517335 275.00
Check your price