Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  70  80  90  100  110  120  130  140  150  155  156  157  158  159  160  161  162  163  164  165  166  167  168  169  170  171  172  173  174  175  176  177  178  179  180  181  182  183  184  190  200  210  220  230  240  250  260  300  400  500  600  700  800  900  1000  1100  1630 
5071 - 5100 of 48877 products displayed
  • F(ab')2-Goat anti-Human IgG Fc Cross-Adsorbed Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # A24480
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24480 147.00
Check your price
  • Goat anti-Rat IgG2a Secondary Antibody, HRP conjugate for Western Blot and ELISA
  • Host Goat
  • Target Species Rat
  • Conjugate HRP
  • Catalog # PA184709
  • Size 500µg
Applications
ELISA (1:10,000), Western Blot (1:1000-1:10000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184709 368.00
Check your price
  • Goat anti-Mouse IgG (Lambda light chain) Secondary Antibody, AP conjugate for ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate AP
  • Catalog # PA186022
  • Size 1mL
Applications
ELISA (1/1000-1/2000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186022 462.00
Check your price
  • CD22 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527928
  • Size 100µL
Applications
Western Blot (1:1000-1:10,000)
Alternate names:
T-cell surface antigen Leu-14; Sialic acid-binding Ig-like lectin 2; B-lymphocyte cell adhesion molecule
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527928 349.00
Check your price
  • Phospho-p70 S6 Kinase (Ser418) Polyclonal Antibody for DB
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512782
  • Size 400µL
Applications
Dot blot (1:500)
Alternate names:
p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I; 70 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512782 349.00
Check your price
  • COX4 Polyclonal Antibody for Western Blot, IHC (P), Flow and IP
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517511
  • Size 100µL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (1:2400), Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
Cytochrome c oxidase subunit IV isoform 1; Cytochrome c oxidase polypeptide IV
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517511 275.00
Check your price
  • IL-33 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA523640
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
C9orf26, DVS27, IL1F11, NF-HEV, NFEHEV, RP11-575C20.2, DVS27-related protein, IL-1F11, interleukin-1 family member 11, interleukin-33, nuclear factor for high endothelial venules, nuclear factor from high endothelial venules
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523640 309.00
Check your price
  • Phospho-SMAD3 (Ser423, Ser425) Monoclonal Antibody for Western Blot, IP and ChIP
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate, Rat, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA514936
  • Size 100µL
Applications
ChIP assay (1:100), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
SMAD family member 3; JV15-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514936 309.00
Check your price
  • MOG Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518351
  • Size 100ug
Applications
Western Blot (0.03-1 ug/ml)
Alternate names:
MOG; MGC26137; myelin oligodendrocyte glycoprotein; DAQB-92E24.2; MOGIG-2; MOG Ig-AluB; MOG alpha-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518351 349.00
Check your price
  • CD9 Monoclonal Antibody, Biotin conjugate for Western Blot and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA119485
  • Size 100µg
Applications
Flow Cytometry (1:400), Western Blot (Assay dependent)
Alternate names:
p24; Tetraspanin-29; Motility-related protein; Leukocyte antigen MIC3; Cell growth-inhibiting gene 2 protein; 5H9 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119485 275.00
Check your price
  • GPR65 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533755
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml)
Alternate names:
T-cell death-associated gene 8 protein; G-protein coupled receptor 65
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533755 349.00
Check your price
  • HAPLN3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523163
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
UNQ238/PRO271, EXLD1, HsT19883, extracellular link domain containing, 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523163 349.00
Check your price
  • ZCCHC4 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531576
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
HSPC052, DHHC domain-containing zinc finger protein, zinc finger CCHC domain-containing protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531576 349.00
Check your price
  • GREM1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513123
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Increased in high glucose protein 2; Down-regulated in Mos-transformed cells protein; DAN domain family member 2; Cysteine knot superfamily 1, BMP antagonist 1; Cell proliferation-inducing gene 2 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513123 349.00
Check your price
  • c-Kit Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA112193
  • Size 100Tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog; p145 c-kit; Tyrosine-protein kinase Kit; Proto-oncogene c-Kit; Piebald trait protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112193 349.00
Check your price
  • Goat anti-Human IgM Heavy Chain Cross-Adsorbed Secondary Antibody, Alexa Fluor® 647 conjugate for IF, ICC and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Alexa Fluor® 647
  • Catalog # A21249
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21249 149.00
Check your price
  • APOE Monoclonal Antibody for Western Blot and IHC (F)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA125289
  • Size 1mL
Applications
Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
B2G1, BG
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125289 349.00
Check your price
  • PPP6C Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515607
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Serine/threonine protein phosphatase 6; PP6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515607 349.00
Check your price
  • Phospho-ErbB4 (Tyr1162) Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512606
  • Size 400µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
p180erbB4; Tyrosine kinase-type cell surface receptor HER4; Proto-oncogene-like protein c-ErbB-4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512606 349.00
Check your price
  • Goat anti-Mouse IgG (H+L) Secondary Antibody, AP conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate AP
  • Catalog # 31321
  • Size 0.2mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:50,000), Immunofluorescence (1:5,000-1:50,000), Immunohistochemistry (1:5,000-1:50,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:50,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31321 105.00
Check your price
  • DHX9 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA541259
  • Size 100µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (0.2-1 µg/ml)
Alternate names:
Nuclear DNA helicase II; DEAH box protein 9; Leukophysin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541259 309.00
Check your price
  • ZNF281 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528290
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Zinc finger DNA-binding protein 99; Transcription factor ZBP-99; GC-box-binding zinc finger protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528290 349.00
Check your price
  • IDH1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520976
  • Size 100ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Oxalosuccinate decarboxylase; NADP(+)-specific ICDH; IDP; Cytosolic NADP-isocitrate dehydrogenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520976 349.00
Check your price
  • G-CSF Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121362
  • Size 250µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Pluripoietin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121362 375.00
Check your price
  • CD5 Monoclonal Antibody, Biotin conjugate for IHC (F), IHC (P), Flow and IP
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA517782
  • Size 100ug
Applications
Flow Cytometry (0.2 ug/10^6 cells), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
Lymphocyte antigen 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517782 129.00
Check your price
  • Annexin A10 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA521297
  • Size 100µg
Applications
ELISA (Assay-Dependent)
Alternate names:
Annexin-14; Annexin-10
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA521297 309.00
Check your price
  • IFN gamma Polyclonal Antibody for Western Blot, IF, ICC and Neu
  • Host Goat
  • Target Species Dog
  • Conjugate Unconjugated
  • Catalog # PA547943
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Neutralization (0.2-1 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
IFN-gamma, IFN-y, Interferon y, IFNG
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547943 349.00
Check your price
  • STX1A Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA516403
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Neuron-specific antigen HPC-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516403 349.00
Check your price
  • Perforin Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512469
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:10)
Alternate names:
Lymphocyte pore-forming protein; Cytolysin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512469 349.00
Check your price
  • MSH3 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529829
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Mismatch repair protein 1; Divergent upstream protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529829 349.00
Check your price