Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  80  90  100  110  120  130  140  150  160  168  169  170  171  172  173  174  175  176  177  178  179  180  181  182  183  184  185  186  187  188  189  190  191  192  193  194  195  196  197  200  210  220  230  240  250  260  270  280  300  400  500  600  700  800  900  1000  1100  1606 
5461 - 5490 of 48165 products displayed
  • Factor X Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513688
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Stuart-Prower factor; Stuart factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513688 349.00
Check your price
  • p300 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515444
  • Size 100µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
E1A-associated protein p300
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515444 349.00
Check your price
  • CPT2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA512217
  • Size 400µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:1000)
Alternate names:
Carnitine palmitoyltransferase II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512217 349.00
Check your price
  • CCL3 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532496
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Tonsillar lymphocyte LD78 alpha protein; Small-inducible cytokine A3; SIS-beta; PAT 464.1; Macrophage inflammatory protein 1-alpha; G0/G1 switch regulatory protein 19-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532496 349.00
Check your price
  • Goat anti-Mouse IgG2a Cross-Adsorbed Secondary Antibody, FITC conjugate for IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # 31634
  • Size 500ug
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:50-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31634 173.00
Check your price
  • Goat anti-Rat IgG Fc Secondary Antibody for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # 31226
  • Size 2mg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31226 119.00
Check your price
  • Plasma Cell Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513532
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513532 309.00
Check your price
  • Adenovirus Type 2/5 Monoclonal Antibody for Western Blot, IF, IHC (P) and IP
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA511349
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1 µg/mL)
Alternate names:
HADV, AdV, Adeno Virus, HADV-2, AdV-2, Adeno Virus 2, HADV-5, AdV-5, Adeno Virus 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511349 309.00
Check your price
  • N-cadherin Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Bovine, Fruit fly, Human, Mouse, Primate, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA517526
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:25), Western Blot (1:1000)
Alternate names:
Neural cadherin; ZNCAD; CDw325
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517526 275.00
Check your price
  • TCR V beta 5 Monoclonal Antibody for Western Blot, Flow, IP and FN
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119020
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Functional Assay (Assay-Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Vbeta5 related, T Cell Receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119020 225.00
Check your price
  • NEU1 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524313
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (2 µg/ml)
Alternate names:
N-acetyl-alpha-neuraminidase 1; Lysosomal sialidase; G9 sialidase; Acetylneuraminyl hydrolase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524313 275.00
Check your price
  • CD7 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MHCD07044
  • Size 2ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
TP41; T-cell surface antigen Leu-9; T-cell leukemia antigen; GP40
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD07044 842.00
Check your price
  • DR4 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA119757
  • Size 0.1mg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
TNF-related apoptosis-inducing ligand receptor 1; Death receptor 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119757 309.00
Check your price
  • ENSA Polyclonal Antibody for Western Blot
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA125716
  • Size 50µg
Applications
Western Blot (1:1,000)
Alternate names:
ARPP-19e
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125716 425.00
Check your price
  • SOX13 Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521556
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
Type 1 diabetes autoantigen ICA12; SRY (Sex determining region Y)-box 13; Islet cell antigen 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521556 349.00
Check your price
  • GPR146 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533635
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (25 µg/ml)
Alternate names:
G-protein coupled receptor PGR8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533635 349.00
Check your price
  • PSMB10 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA544572
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Proteasome subunit beta-2i; Proteasome MECl-1; Multicatalytic endopeptidase complex subunit MECl-1; Macropain subunit MECl-1; Low molecular mass protein 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544572 309.00
Check your price
  • FGFR5 Polyclonal Antibody for Western Blot and IHC (F)
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547218
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Fibroblast growth factor receptor 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547218 349.00
Check your price
  • FANCD2 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA123333
  • Size 100µg
Applications
ELISA (Assay dependent), Western Blot (1:500-1:100)
Alternate names:
DKFZp762A223, FA-D2, FA4, FACD, FAD, FAD2, FANCD, FLJ23826
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123333 309.00
Check your price
  • Goat anti-Human IgA Cross-Adsorbed Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate HRP
  • Catalog # A18787
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18787 149.00
Check your price
  • PrP Monoclonal Antibody for IHC (P) and ELISA
  • Host Mouse
  • Target Species Sheep
  • Conjugate Unconjugated
  • Catalog # MA182828
  • Size 250µg
Applications
ELISA (1:50-1:1000), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
ASCR, CD230, CJD, GSS, MGC26679, PRIP, PrP, PrP27-30, PrP33-35C, PrPc, PRNP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182828 375.00
Check your price
  • OVOL2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531665
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Zinc finger protein 339
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531665 349.00
Check your price
  • F(ab')2-Goat anti-Rabbit IgG (H+L) Secondary Antibody, Qdot® 655 conjugate for Western Blot, ICC, IHC and Flow
  • Host Goat
  • Target Species Rabbit
  • Conjugate Qdot® 655
  • Catalog # Q11421MP
  • Size 200µL
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50), Immunohistochemistry (1:50), Western Blot (1:50)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
Q11421MP 319.00
Check your price
  • LIN28A Monoclonal Antibody, HRP conjugate for Western Blot
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate HRP
  • Catalog # MA1016HRP
  • Size 100µL
Applications
Western Blot (1:250-1:1000 )
Alternate names:
Testis-expressed protein 17; Zinc finger CCHC domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1016HRP 275.00
Check your price
  • MESP2 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517121
  • Size 100ug
Applications
Western Blot (1:500-1:2000)
Alternate names:
Class C basic helix-loop-helix protein 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517121 309.00
Check your price
  • Annexin A4 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA547586
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Annexin-4; Annexin IV; Protein II; Placental anticoagulant protein II; PP4-X; P32.5; Lipocortin IV; Endonexin I; Chromobindin-4; Carbohydrate-binding protein p33/p41; 35-beta calcimedin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547586 375.00
Check your price
  • Donkey anti-Sheep IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 680 conjugate for Western Blot, IF and ICC
  • Host Donkey
  • Target Species Sheep
  • Conjugate Alexa Fluor® 680
  • Catalog # A21102
  • Size 500µL
Applications
Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL), Western Blot (0.04-0.2 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21102 209.00
Check your price
  • VPS4B Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530316
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Suppressor of K(+) transport growth defect 1; Cell migration-inducing gene 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530316 349.00
Check your price
  • F(ab')2-Goat anti-Human IgG (H+L) Cross-Adsorbed Secondary Antibody, TRITC conjugate for IF
  • Host Goat
  • Target Species Human
  • Conjugate TRITC
  • Catalog # A24475
  • Size 500µg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24475 118.00
Check your price
  • Listeria monocytogenes Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Eubacteria
  • Conjugate Unconjugated
  • Catalog # MA120269
  • Size 100µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
L. monocytogenes
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120269 309.00
Check your price