Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  80  90  100  110  120  130  140  150  160  169  170  171  172  173  174  175  176  177  178  179  180  181  182  183  184  185  186  187  188  189  190  191  192  193  194  195  196  197  198  200  210  220  230  240  250  260  270  280  300  400  500  600  700  800  900  1000  1100  1622 
5491 - 5520 of 48655 products displayed
  • Actin Muscle Monoclonal Antibody for Western Blot, IF, IHC (P) and Flow
  • Host Mouse
  • Target Species Bovine, Dog, Horse, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA511874
  • Size 500 µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1-2 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin-1; Alpha-cardiac actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511874 249.00
Check your price
  • F(ab')2-Goat anti-Human IgG Secondary Antibody, PE conjugate for Flow
  • Host Goat
  • Target Species Human
  • Conjugate PE
  • Catalog # PA184341
  • Size 1 mL
Applications
Flow Cytometry (Neat-1/10)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184341 397.00
Check your price
  • CHERP Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse, Rat, Zebrafish, Bovine, Dog, Horse, Guinea pig, Human
  • Conjugate Unconjugated
  • Catalog # PA541813
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
SR-related CTD-associated factor 6; ERPROT 213-21
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541813 309.00
Check your price
  • STING Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523337
  • Size 100 ug
Applications
Western Blot (5-10 µg/ml)
Alternate names:
Transmembrane protein 173; Mediator of IRF3 activation; Endoplasmic reticulum interferon stimulator
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523337 349.00
Check your price
  • GKAP1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA525388
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
cGMP-dependent protein kinase-anchoring protein of 42 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525388 349.00
Check your price
  • MMP14 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532500
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:30)
Alternate names:
Membrane-type-1 matrix metalloproteinase; Membrane-type matrix metalloproteinase 1; MMP-X1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532500 275.00
Check your price
  • SERCA2 ATPase Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529380
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Endoplasmic reticulum class 1/2 Ca(2+) ATPase; Calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletal muscle isoform; Calcium pump 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529380 349.00
Check your price
  • Cryptosporidium parvum Oocysts Monoclonal Antibody for IF, ICC and ELISA
  • Host Mouse
  • Target Species Protozoa
  • Conjugate Unconjugated
  • Catalog # MA518040
  • Size 100 µL
Applications
ELISA (Assay-dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent)
Alternate names:
Cryptosporidium parvum, Cryptosporidiosis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518040 425.00
Check your price
  • LDHA Monoclonal Antibody for Western Blot, Flow, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA517247
  • Size 100 µL
Applications
ELISA (Assay Dependent), Immunoprecipitation (2 µg), Western Blot (0.1 µg/ml)
Alternate names:
Renal carcinoma antigen NY-REN-59; LDH muscle subunit; Cell proliferation-inducing gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517247 349.00
Check your price
  • JPH1 Polyclonal Antibody for Western Blot, IF, IHC (F), IHC (P) and Flow
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 405100
  • Size 100 µg
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Junctophilin type 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
405100 305.00
Check your price
  • MMP13 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA516566
  • Size 500 µL
Applications
Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Western Blot (2.5-5.0 µg/ml)
Alternate names:
Matrix metalloproteinase-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516566 309.00
Check your price
  • beta Catenin Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515569
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515569 349.00
Check your price
  • E. coli serotype K99 Pili Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA17031
  • Size 1 mL
Applications
ELISA (1:20)
Alternate names:
Escherichia coli
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17031 349.00
Check your price
  • SERPINA3 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110630
  • Size 500 µg
Applications
ELISA (Assay dependent)
Alternate names:
Serpin A3; Cell growth-inhibiting gene 24/25 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110630 349.00
Check your price
  • HPx2 Monoclonal Antibody for IF, ICC, IHC (F) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516133
  • Size 100 µL
Applications
Flow Cytometry (1:50-1:100), Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Frozen) (1:100)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516133 349.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 546 conjugate for IF, ICC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 546
  • Catalog # A11003
  • Size 500 µL
Applications
Flow Cytometry (1-10 µg/mL), Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11003 169.00
Check your price
  • Phospho-c-Abl (Tyr245) Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA512532
  • Size 400 µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
p150; Proto-oncogene c-Abl; Abelson tyrosine-protein kinase 1; Abelson murine leukemia viral oncogene homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512532 349.00
Check your price
  • PRDM6 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA543659
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
PR domain-containing protein 6; PR domain zinc finger protein 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543659 309.00
Check your price
  • EphA10 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515848
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
EphA10s protein; ephrin type-A receptor 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515848 349.00
Check your price
  • HA Tag Monoclonal Antibody, DyLight® 680 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate DyLight® 680
  • Catalog # 26183D680
  • Size 50 µL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (1:500-1:1000)
Alternate names:
human influenza hemagglutinin, HA-tag, HA, HA Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
26183D680 159.00
Check your price
  • Phalloidin, DyLight® 594 conjugate for IF, ICC and IHC
  • Host
  • Target Species Many
  • Conjugate DyLight® 594
  • Catalog # 21836
  • Size 300 units
Applications
Immunocytochemistry (1-5 units/ml), Immunofluorescence (1-5 units/ml), Immunohistochemistry (Assay dependent)
Alternate names:
DY594-Phalloidin, F-actin toxin, Amanita phalloides
Catalog # Size Price (AUD) Your Price (AUD) Qty
21836 378.00
Check your price
  • FXR Monoclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 417200
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Retinoid X receptor-interacting protein 14; Nuclear receptor subfamily 1 group H member 4; Farnesol receptor HRR-1; Farnesoid X-activated receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
417200 427.00
Check your price
  • CD8 alpha Monoclonal Antibody, PE-Cyanine7 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE-Cyanine7
  • Catalog # A15846
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
T-lymphocyte differentiation antigen T8/Leu-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15846 Not Orderable Online
Check your price
  • Mast Cell Chymase Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511717
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:2000), Western Blot (Assay Dependent)
Alternate names:
Mast cell protease I; Alpha-chymase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511717 309.00
Check your price
  • CD2/CD20 Monoclonal Antibody, FITC, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC , PE
  • Catalog # MA112058
  • Size 100 Tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Membrane-spanning 4-domains subfamily A member 1; Leukocyte surface antigen Leu-16; Bp35; B-lymphocyte surface antigen B1; T-cell surface antigen T11/Leu-5; Rosette receptor; LFA-3 receptor; LFA-2; Erythrocyte receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112058 375.00
Check your price
  • Adenovirus Type 5 Hexon Polyclonal Antibody, HRP conjugate for ELISA
  • Host Goat
  • Target Species Virus
  • Conjugate HRP
  • Catalog # PA173054
  • Size 1 mL
Applications
ELISA (1:200-1:1,000)
Alternate names:
HADV, AdV, Adeno Virus, HADV-5, AdV-5, Adeno Virus 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173054 375.00
Check your price
  • CDw17 Monoclonal Antibody for IF, ICC and Flow
  • Host Mouse
  • Target Species Human, Many
  • Conjugate Unconjugated
  • Catalog # MA191271
  • Size 200 µg
Applications
Flow Cytometry (1-5 µg for 10^6 cells), Immunocytochemistry (5-20 ug/ml), Immunofluorescence (5-20 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191271 349.00
Check your price
  • GCP5 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539182
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Gamma-tubulin complex component 5, GCP-5, TUBGCP5, GCP5, KIAA1899
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539182 309.00
Check your price
  • Collagen IV Polyclonal Antibody for IF, IHC (P) and ELISA
  • Host Rabbit
  • Target Species Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA185320
  • Size 100 µL
Applications
ELISA (1:100-1:400), Immunofluorescence (1:40-1:160), Immunohistochemistry (Paraffin) (1:250-1:1000)
Alternate names:
arresten
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185320 425.00
Check your price
  • CCDC47 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520855
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Adipocyte-specific protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520855 349.00
Check your price