Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  31  32  33  40  50  60  70  80  90  100  110  200  300  400  500  600  700  800  900  1000  1611 
541 - 570 of 48310 products displayed
  • Goat anti-Mouse IgG (H+L), Alexa Fluor® 488
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 488
  • Catalog # A28175
  • Size 1 mg
Applications
Flow Cytometry (1-5 µg/ml), Immunocytochemistry (0.1-2 µg/ml), Immunofluorescence (0.1-2 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A28175 249.00
Check your price
  • CaMKII alpha Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # A14012
  • Size 100 µL
Applications
Western Blot (1:1,000)
Alternate names:
calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, calcium/calmodulin-dependent protein kinase type II alpha chain, Calci
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14012 309.00
Check your price
  • CD200R Antibody (Monoclonal, OX-102)
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA516894
  • Size 250 ug
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
OX102 antigen; Cell surface glycoprotein OX2 receptor 1; CD200 cell surface glycoprotein receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516894 309.00
Check your price
  • Troponin I Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Goat, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA542108
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (4-8 µg/ml), Western Blot (0.2-1 µg/ml)
Alternate names:
Troponin I, slow-twitch isoform
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542108 309.00
Check your price
  • CD45 Antibody, APC-Alexa Fluor® 750 (Monoclonal, HI30)
  • Host Mouse
  • Target Species Human
  • Conjugate APC-Alexa Fluor® 750
  • Catalog # MHCD4527
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
T200; Leukocyte common antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD4527 332.00
Check your price
  • Phospho-LATS1/LATS2 (Thr1079, Thr1041) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA537834
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Serine/threonine-protein kinase kpm; Large tumor suppressor homolog 2; Kinase phosphorylated during mitosis protein; WARTS protein kinase; Large tumor suppressor homolog 1; Warts-like kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537834 309.00
Check your price
  • CD16 Antibody, PE (Monoclonal, MEM-154)
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA119631
  • Size 100 tests
Applications
Flow Cytometry (20ul x 100ul whole blood or 10^6 cells)
Alternate names:
IgG Fc receptor III-2; FcR-10; Fc-gamma RIII-alpha; CD16a antigen; IgG Fc receptor III-1; Fc-gamma RIII-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119631 349.00
Check your price
  • VEGF Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA516754
  • Size 500 µL
Applications
Immunocytochemistry (1:40-1:200), Immunofluorescence (1:40-1:200), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (3 µg/ml), Western Blot (1 µg/mL)
Alternate names:
Vascular permeability factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516754 309.00
Check your price
  • Goat anti-Mouse IgG Fc Cross-Adsorbed, PE
  • Host Goat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # 31861
  • Size 1 mL
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:50-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31861 232.00
Check your price
  • Progesterone Receptor Antibody (Monoclonal, PgR636)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512581
  • Size 500 µL
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:10-1:20), Western Blot (1:500-1:1000)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512581 850.00
Check your price
  • IGF1R alpha Antibody (Monoclonal, 17-69)
  • Host Mouse
  • Target Species Human, Pig, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA513807
  • Size 500 µL
Applications
Immunoprecipitation (2 µg/ml), Inhibition Assays (Assay Dependent)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513807 349.00
Check your price
  • 6x-His Tag Antibody, Alexa Fluor® 647 (Monoclonal, His.H8)
  • Host Mouse
  • Target Species Tag
  • Conjugate Alexa Fluor® 647
  • Catalog # MA121315A647
  • Size 50 µl
Applications
Western Blot (1:500)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121315A647 225.00
Check your price
  • Goat anti-Mouse IgG (H+L), HRP
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # 626520
  • Size 1 mL
Applications
ELISA (1:2,000-1:4,000), Immunohistochemistry (1:2,000-1:4,000), Western Blot (1:2,000-1:4,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
626520 149.00
Check your price
  • Mouse IgG1, PE Isotype Control
  • Host Mouse
  • Target Species Not Applicable
  • Conjugate PE
  • Catalog # MA110415
  • Size 100 ug
Applications
Control (Assay-Dependent), Flow Cytometry (Assay Dependent)
Alternate names:
Negative Control, Immunoglobulin Control, Control
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110415 228.00
Check your price
  • MUC3 Antibody (Monoclonal, M3.1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135702
  • Size 100 µg
Applications
ELISA (Assay dependent.), Flow Cytometry (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.), Immunoprecipitation (Assay dependent.), Western Blot (Assay dependent.)
Alternate names:
Mucin-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135702 309.00
Check your price
  • IP3 Receptor 2 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA541009
  • Size 100 µl
Applications
Western Blot (1 µg/ml )
Alternate names:
Type 2 inositol 1,4,5-trisphosphate receptor; Inositol 1,4,5-trisphosphate type V receptor; IP3 receptor isoform 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541009 309.00
Check your price
  • SENP7 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536089
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Sentrin/SUMO-specific protease SENP7; SUMO-1-specific protease 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536089 309.00
Check your price
  • Goat anti-Mouse IgA Cross-Adsorbed
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 626700
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
626700 118.00
Check your price
  • VE-cadherin Antibody
  • Host Rabbit
  • Target Species Chicken, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519612
  • Size 100 µg
Applications
Flow Cytometry (Assay-Dependent), Immunocytochemistry (5 µg/ml), Immunofluorescence (5 µg/ml), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1 µg/ml)
Alternate names:
Vascular endothelial cadherin; 7B4 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519612 349.00
Check your price
  • MARCO Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526888
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Scavenger receptor class A member 2; Macrophage receptor with collagenous structure
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526888 349.00
Check your price
  • M13 Phage coat protein Antibody
  • Host Rabbit
  • Target Species Bacteria, Virus
  • Conjugate Unconjugated
  • Catalog # PA126758
  • Size 100 µL
Applications
ELISA (1/1000), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126758 399.00
Check your price
  • ARF6 Antibody (Monoclonal, 6ARF01)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA511795
  • Size 500 µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Western Blot (1:100-1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511795 349.00
Check your price
  • Human Stem Cell Factor (SCF) Recombinant Protein
  • Host E. coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP8631
  • Size 10 µg
Applications
Control (Assay Dependent)
Alternate names:
c-Kit ligand; Stem cell factor; Mast cell growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP8631 265.00
Check your price
  • Integrin beta 7 Antibody (Monoclonal, 473207)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524192
  • Size 100 µg
Applications
Flow Cytometry (0.25 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml)
Alternate names:
Gut homing receptor beta subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524192 275.00
Check your price
  • Cytokeratin Pan Antibody (Monoclonal, C11)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA112594
  • Size 100 µg
Applications
Immunohistochemistry (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p53-related protein; p53-like transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112594 375.00
Check your price
  • Bacillus anthracis Protective Antigen Antibody (Monoclonal, BAP0101)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA121675
  • Size 100 µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
B. anthracis PA, Bacillus anthracis PA, B. anthracis Protective Antigen, Anthrax Protective Antigen, Anthrax PA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121675 349.00
Check your price
  • IL11RA Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539583
  • Size 100 µg
Applications
Western Blot (1:500-1:3000)
Alternate names:
Soluble form, sIL11RA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539583 309.00
Check your price
  • LDLR Antibody (Monoclonal, 472413)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523916
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
FH, FHC, LDLCQ2, LDL receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523916 275.00
Check your price
  • Chlamydia trachomatis Antibody (Monoclonal, A77C)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA17336
  • Size 100 µg
Applications
ELISA (1:20), Immunofluorescence (1:10), Immunohistochemistry (1:10)
Alternate names:
C. trachomatis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17336 309.00
Check your price
  • Desmocollin 2 Antibody
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547651
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Desmosomal glycoprotein III; Desmosomal glycoprotein II; Desmocollin-3; Cadherin family member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547651 349.00
Check your price