Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  90  100  110  120  130  140  150  160  170  182  183  184  185  186  187  188  189  190  191  192  193  194  195  196  197  198  199  200  201  202  203  204  205  206  207  208  209  210  211  220  230  240  250  260  270  280  290  300  400  500  600  700  800  900  1000  1100  1630 
5881 - 5910 of 48877 products displayed
  • Cardiac Troponin T Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human, Pig
  • Conjugate Unconjugated
  • Catalog # MA124620
  • Size 200µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Cardiac muscle troponin T
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124620 309.00
Check your price
  • ELF3 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521293
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Epithelium-specific Ets transcription factor 1; Epithelium-restricted Ets protein ESX; Epithelial-restricted with serine box; E74-like factor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521293 349.00
Check your price
  • HOXB9 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA540576
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (4-8 µg/ml), Western Blot (0.2-1 µg/ml)
Alternate names:
Homeobox protein Hox-2.5; Homeobox protein Hox-2E
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540576 309.00
Check your price
  • PABPN1 Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Bovine, Dog, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA541793
  • Size 100µl
Applications
Immunocytochemistry (1:200), Western Blot (1.25 µg/ml)
Alternate names:
Polyadenylate-binding nuclear protein 1; Poly(A)-binding protein II; Nuclear poly(A)-binding protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541793 309.00
Check your price
  • ITGA7 Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA523653
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Integrin alpha-7, Integrin alpha-7 light chain, Integrin alpha-7 70 kDa form, ITGA7, UNQ406/PRO768
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523653 339.00
Check your price
  • CD163 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate PE
  • Catalog # MA516657
  • Size 100TESTS
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
CD163 antigen, hemoglobin scavenger receptor, macrophage-associated antigen, scavenger receptor cysteine-rich type 1 protein M130
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516657 650.00
Check your price
  • FPR1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533536
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml)
Alternate names:
N-formylpeptide chemoattractant receptor; N-formyl peptide receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533536 349.00
Check your price
  • GFAP Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Chicken, Guinea pig, Hamster, Human, Mouse, Non-human primate, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # PA516291
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516291 299.00
Check your price
  • Bovine RSV Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA123355
  • Size 100µg
Applications
ELISA (Assay dependent), Western Blot (1:500-1:1000)
Alternate names:
BRSV
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123355 375.00
Check your price
  • cAMP Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA515393
  • Size 100µL
Applications
ELISA (1:10,000)
Alternate names:
Cyclic AMP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515393 349.00
Check your price
  • Claudin 1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA532350
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:200), Western Blot (1:25)
Alternate names:
Senescence-associated epithelial membrane protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532350 309.00
Check your price
  • V5 Tag Monoclonal Antibody for Western Blot, IF, ICC, Flow, IP and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # R96025
  • Size 50µl
Applications
ELISA (Assay dependent), Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunoprecipitation (Assay Dependent), Western Blot (1:1000-1:5000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
R96025 196.00
Check your price
  • SMYD5 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513217
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Retinoic acid-induced protein 15; Protein NN8-4AG
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513217 349.00
Check your price
  • RAB11 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA124919
  • Size 100µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
YL8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124919 375.00
Check your price
  • CD45 Monoclonal Antibody, APC-Cyanine7 conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate APC-Cyanine7
  • Catalog # A15395
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
B220, CD45, CD45R, GP180, L-CA, LCA, LY5, T200, CD45 antigen, T200 glycoprotein, T200 leukocyte common antigen, protein tyrosine phosphatase, receptor type, c polypeptide, receptor-type tyrosine-protein phosphatase C
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15395 204.00
Check your price
  • Factor X Monoclonal Antibody for Western Blot and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA143035
  • Size 100µg
Applications
ELISA (Assay Dependent.), Western Blot (5 ug/ml)
Alternate names:
Stuart factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA143035 309.00
Check your price
  • P2Y6 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534093
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (17-34 µg/ml)
Alternate names:
P2RY6, G-coupled nucleotide receptor, p2 purinoceptor, p2y3 receptor, p2Y purinoceptor 6, Purinergic receptor p2y6, p2RY6, p2y3 and chick, p2Y6 receptor, p2Y6, Purinoceptor p2y6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534093 349.00
Check your price
  • SPIN1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536363
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Spindlin1; Ovarian cancer-related protein; SSEC P; 30000 Mr metaphase complex
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536363 309.00
Check your price
  • F(ab')2-Goat anti-Human IgG (H+L) Secondary Antibody, Qdot® 655 conjugate for Western Blot, ICC, IHC and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Qdot® 655
  • Catalog # Q11221MP
  • Size 200µL
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50), Immunohistochemistry (1:50), Western Blot (1:50)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
Q11221MP 319.00
Check your price
  • JunD Polyclonal Antibody for Western Blot, IF, ICC, Flow and ChIP
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # 720035
  • Size 100µg
Applications
ChIP assay (2-4 µg x 10^6 cells), Flow Cytometry (2-4 µg x 10^6 cells), Immunocytochemistry (2 µg/ml), Immunofluorescence (2 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Transcription factor jun-D, Oncogene Jun-D
Catalog # Size Price (AUD) Your Price (AUD) Qty
720035 325.00
Check your price
  • Rabbit IgG Control Isotype Control for Flow, Ctrl
  • Host Rabbit
  • Target Species Not Applicable
  • Conjugate
  • Catalog # PA523093
  • Size 100ug
Applications
Control (Assay-Dependent), Flow Cytometry (1 µl/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523093 Not Orderable Online
Check your price
  • Eotaxin 2 Polyclonal Antibody for Western Blot, ELISA, Neu, FN and IA
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA125517
  • Size 50µg
Applications
ELISA (0.5 µg/ml), Functional Assay (7-8 ug/ml), Inhibition Assays (7-8 ug/ml), Neutralization (7-8 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
Small-inducible cytokine A24; Eotaxin-2; Eosinophil chemotactic protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125517 375.00
Check your price
  • alpha-1 Antitrypsin Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mammal
  • Conjugate Unconjugated
  • Catalog # PA122860
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:25-1:50)
Alternate names:
Serpin A1; Alpha-1-antiproteinase; Alpha-1 protease inhibitor; Alpha-1-proteinase inhibitor; Alpha-1-antitrypsin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA122860 309.00
Check your price
  • HSD3B1 Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Bovine, Human, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA146438
  • Size 100µL
Applications
Immunocytochemistry (10-20 µg/ml), Immunofluorescence (10-20 µg/ml), Immunohistochemistry (Frozen) (10-20 µg/ml), Immunohistochemistry (Paraffin) (10-20 µg/ml), Western Blot (Assay-Dependent)
Alternate names:
Trophoblast antigen FDO161G; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type I
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146438 309.00
Check your price
  • SLC27A5 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA542028
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Very long-chain acyl-CoA synthetase-related protein; Solute carrier family 27 member 5; Fatty acid transport protein 5; Cholate--CoA ligase; Bile acid-CoA ligase; Very long-chain acyl-CoA synthetase homolog 2; Fatty-acid-coenzyme A ligase, very long-chain 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542028 309.00
Check your price
  • SMN Monoclonal Antibody for Western Blot, IHC and Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA515857
  • Size 100µL
Applications
Flow Cytometry (1/200-1/400), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Gemin-1; Component of gems 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515857 349.00
Check your price
  • Hemoglobin Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIH9505
  • Size 1mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Hemoglobin alpha chain; Alpha-globin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9505 600.00
Check your price
  • NR1D2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530342
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
V-erbA-related protein 1-related; Rev-erb-beta; Rev-erb alpha-related receptor; Orphan nuclear hormone receptor BD73
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530342 349.00
Check your price
  • RAB33B Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530697
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
ras-related protein Rab-33B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530697 349.00
Check your price
  • CXCL1 Monoclonal Antibody for Western Blot, Flow, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523707
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Neutralization (3-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Neutrophil-activating protein 3; Melanoma growth stimulatory activity; GRO-alpha(1-73); C-X-C motif chemokine 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523707 375.00
Check your price