Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  100  110  120  130  140  150  160  170  180  193  194  195  196  197  198  199  200  201  202  203  204  205  206  207  208  209  210  211  212  213  214  215  216  217  218  219  220  221  222  230  240  250  260  270  280  290  300  400  500  600  700  800  900  1000  1100  1200  1630 
6211 - 6240 of 48882 products displayed
  • Phospho-c-Jun (Ser63) Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA517890
  • Size 100µL
Applications
Flow Cytometry (1:100), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
V-jun avian sarcoma virus 17 oncogene homolog; Proto-oncogene c-Jun; Activator protein 1; AH119; p39
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517890 309.00
Check your price
  • Protein C Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528321
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Blood coagulation factor XIV; Autoprothrombin IIA; Anticoagulant protein C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528321 349.00
Check your price
  • RORC Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534166
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (22 µg/ml)
Alternate names:
Retinoid-related orphan receptor-gamma; RAR-related orphan receptor C; Nuclear receptor subfamily 1 group F member 3; Nuclear receptor RZR-gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534166 349.00
Check your price
  • PSA Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517108
  • Size 100µL
Applications
Flow Cytometry (1:200-1:400), Western Blot (1:500-1:2000)
Alternate names:
Semenogelase; P-30 antigen; Kallikrein-3; Gamma-seminoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517108 309.00
Check your price
  • Phospho-OPRD1 (Ser363) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538252
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Opioid receptor A; MSL-2; K56
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538252 309.00
Check your price
  • UBE2N Monoclonal Antibody for Western Blot, IF and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 371100
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Ubiquitin-protein ligase N; Ubiquitin carrier protein N; E2 ubiquitin-conjugating enzyme N; Bendless-like ubiquitin-conjugating enzyme; Ubc13; UbcH13
Catalog # Size Price (AUD) Your Price (AUD) Qty
371100 307.00
Check your price
  • CD3e Monoclonal Antibody, APC-Cyanine7 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC-Cyanine7
  • Catalog # A15440
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
T-cell surface antigen T3/Leu-4 epsilon chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15440 289.00
Check your price
  • Phospho-IRS1 (Ser312) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44814G
  • Size 100µL
Applications
Western Blot (Assay Dependent)
Alternate names:
IRS-1, IRS1, Insulin receptor substrate-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44814G 309.00
Check your price
  • PPAR gamma (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP75683
  • Size 100µg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Peroxisome Proliferator-Acivated Receptor Gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75683 588.00
Check your price
  • SNX15 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528459
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
HSAF001435, clone iota unknown protein, sorting nexin-15
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528459 349.00
Check your price
  • Clostridium tetani Tetanus Toxin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # HYB2780102
  • Size 200µl
Applications
ELISA (1:8,000)
Alternate names:
C. tetani, pE88_60
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB2780102 650.00
Check your price
  • PILRA Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547613
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
Inhibitory receptor PILR-alpha; Cell surface receptor FDF03
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547613 349.00
Check your price
  • RIF1 Polyclonal Antibody for Western Blot and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA537951
  • Size 100µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Rap1-interacting factor 1 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537951 309.00
Check your price
  • Donkey anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 680 conjugate for Western Blot, IF and ICC
  • Host Donkey
  • Target Species Goat
  • Conjugate Alexa Fluor® 680
  • Catalog # A21084
  • Size 500µL
Applications
Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL), Western Blot (0.04-0.2 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21084 209.00
Check your price
  • SRC Oligoclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 710449
  • Size 100µg
Applications
Immunocytochemistry (1-2 ug/ml), Immunofluorescence (1-2 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
pp60c-src; Proto-oncogene c-Src
Catalog # Size Price (AUD) Your Price (AUD) Qty
710449 309.00
Check your price
  • DC-SIGN Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Human
  • Conjugate PE
  • Catalog # A18375
  • Size 100tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Dendritic cell-specific ICAM-3-grabbing non-integrin 1; C-type lectin domain family 4 member L
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18375 178.00
Check your price
  • Phospho-S1P1 (Thr236) Polyclonal Antibody for IF, ICC and pep-ELISA
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA538813
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), peptide-ELISA (1:10,000)
Alternate names:
Sphingosine 1-phosphate receptor Edg-1; Endothelial differentiation G-protein coupled receptor 1; Lysophospholipid receptor B1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538813 309.00
Check your price
  • CD45R Monoclonal Antibody, TRI-COLOR® conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate TRI-COLOR®
  • Catalog # RM2606
  • Size 500µL
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
B220, CD45R, Cd45, Ly-5, Lyt-4, T200, Cluster designation 45R
Catalog # Size Price (AUD) Your Price (AUD) Qty
RM2606 258.00
Check your price
  • LIMK1/LIMK2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536062
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
KIZ-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536062 309.00
Check your price
  • STX1A Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA517612
  • Size 200ug
Applications
ELISA (Assay Dependent), Flow Cytometry (0.5 ug/10^6 cells), Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (Paraffin) (1:500), Western Blot (Assay Dependent)
Alternate names:
Neuron-specific antigen HPC-1; Synaptotagmin-associated 35 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517612 325.00
Check your price
  • Progesterone Receptor Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517226
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517226 275.00
Check your price
  • SNAP25 Monoclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA112710
  • Size 100µg
Applications
Immunohistochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Synaptosomal-associated 25 kDa protein; Super protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112710 309.00
Check your price
  • DNA Adenine Methylase Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA112717
  • Size 100µg
Applications
Western Blot (5-10 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112717 375.00
Check your price
  • Septin-7 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521365
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
CDC10 protein homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521365 349.00
Check your price
  • CD105 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MHCD10504
  • Size 0.5ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
CD105, END, HHT1, ORW, ORW1, CD105 antigen, ENG
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD10504 267.00
Check your price
  • SLC33A1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA542441
  • Size 100µl
Applications
Immunocytochemistry (1:600), Immunofluorescence (1:600), Immunohistochemistry (Paraffin) (1:600), Western Blot (0.2-1 µg/ml)
Alternate names:
Solute carrier family 33 member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542441 309.00
Check your price
  • CD45RA Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MHCD45RA04
  • Size 0.5ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
T200; Leukocyte common antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD45RA04 268.00
Check your price
  • FGL2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536390
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Prothrombinase; Fibrinogen-like protein 2; Cytotoxic T-lymphocyte-specific protein; pT49
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536390 309.00
Check your price
  • DNP Monoclonal Antibody for IHC and ELISA
  • Host Rat
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # 048888
  • Size 200µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
048888 333.00
Check your price
  • CRY1 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1527
  • Size 100µg
Applications
Western Blot (1:2,000)
Alternate names:
Cry1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1527 349.00
Check your price