Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  120  130  140  150  160  170  180  190  200  204  205  206  207  208  209  210  211  212  213  214  215  216  217  218  219  220  221  222  223  224  225  226  227  228  229  230  231  232  233  240  250  260  270  280  290  300  310  400  500  600  700  800  900  1000  1100  1200  1630 
6541 - 6570 of 48882 products displayed
  • PPAR alpha Monoclonal Antibody for Western Blot, IF, ICC, Flow, IP, ChIP and GS
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1822
  • Size 100µg
Applications
ChIP assay (Assay Dependent), Flow Cytometry (1µg per 10^6 cells), Gel Shift (Assay dependent), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunoprecipitation (Assay dependent), Western Blot (2-5 µg/ml)
Alternate names:
Nuclear receptor subfamily 1 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1822 375.00
Check your price
  • IL-27 p28 Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710175
  • Size 100µg
Applications
Western Blot (1-3 ug/ml)
Alternate names:
p28; Interleukin-30
Catalog # Size Price (AUD) Your Price (AUD) Qty
710175 309.00
Check your price
  • RSV NP Monoclonal Antibody, FITC conjugate for IF and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate FITC
  • Catalog # MA17289
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunofluorescence (1:5)
Alternate names:
human respiratory syncytial virus, RSV, RSV NP, RSV Nuclear Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17289 349.00
Check your price
  • GGTLC2 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523722
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Gamma-glutamyltransferase-like protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523722 349.00
Check your price
  • IL1R2 Polyclonal Antibody for Western Blot, IF, ICC and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547359
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Neutralization (15-30 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Interleukin-1 receptor type II; Interleukin-1 receptor beta; IL-1 type II receptor; CDw121b; CD121 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547359 349.00
Check your price
  • PCDH17 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA537941
  • Size 100µg
Applications
Western Blot (0.1-0.3 µg/ml)
Alternate names:
protocadherin 17, PCDH68, PCH68, protocadherin 68, protocadherin-17, protocadherin-68
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537941 309.00
Check your price
  • IHH Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519680
  • Size 100µg
Applications
Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
HHG-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519680 349.00
Check your price
  • FAM55D Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA542687
  • Size 100µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (5 µg/ml)
Alternate names:
Protein FAM55D
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542687 309.00
Check your price
  • OTUB1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521402
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Ubiquitin-specific-processing protease OTUB1; Otubain-1; OTU domain-containing ubiquitin aldehyde-binding protein 1; Deubiquitinating enzyme OTUB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521402 349.00
Check your price
  • RAD52 (human) for Western Blot and Ctrl
  • Host Human
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP4973
  • Size 100µg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP4973 1,900.00
Check your price
  • Cytokeratin 10 Monoclonal Antibody, Biotin conjugate for Western Blot, IF and IHC (P)
  • Host Mouse
  • Target Species Dog, Cat, Human, Rat
  • Conjugate Biotin
  • Catalog # MA513702
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (0.5-1.0 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Keratin-10; Cytokeratin-10; Type I keratin Ka10
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513702 349.00
Check your price
  • Factor XI (human) for Ctrl
  • Host Human
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP43075
  • Size 50µg
Applications
Control (Assay Dependent)
Alternate names:
F11
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP43075 159.00
Check your price
  • Smooth Muscle Actin Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515805
  • Size 100µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515805 349.00
Check your price
  • RPRD1B Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA531614
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Cell cycle-related and expression-elevated protein in tumor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531614 349.00
Check your price
  • PEA15 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517326
  • Size 100µL
Applications
Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
15 kDa phosphoprotein enriched in astrocytes; Phosphoprotein enriched in diabetes
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517326 275.00
Check your price
  • HTRA1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523395
  • Size 100µL
Applications
Immunocytochemistry (1:10-1:2000), Immunofluorescence (1:10-1:2000), Western Blot (1:250-1:1000)
Alternate names:
Serine protease 11; High-temperature requirement A serine peptidase 1; L56
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523395 349.00
Check your price
  • Nuclear Membrane Monoclonal Antibody for IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135738
  • Size 100µg
Applications
Flow Cytometry (Assay dependent.), Immunohistochemistry (Paraffin, Frozen) (Assay dependent.)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135738 309.00
Check your price
  • HMGN2 Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538584
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
High mobility group nucleosome-binding domain-containing protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538584 309.00
Check your price
  • IL-33 Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515772
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (1 ug/ml)
Alternate names:
Nuclear factor from high endothelial venules; Interleukin-1 family member 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515772 349.00
Check your price
  • NKp46 Polyclonal Antibody for Western Blot, Flow and FN
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546986
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Functional Assay (0.4-2.4 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Natural killer cell p46-related protein; Lymphocyte antigen 94; Activating receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546986 399.00
Check your price
  • RPN1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA512153
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
Ribophorin-1; Ribophorin I; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase 67 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512153 349.00
Check your price
  • Lactoferrin Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA146413
  • Size 100µg
Applications
ELISA (1:100-1:2000), Western Blot (1:100-1:2000)
Alternate names:
Talalactoferrin; Growth-inhibiting protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146413 309.00
Check your price
  • FAM3C Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA141632
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (0.5-3 µg/ml)
Alternate names:
Interleukin-like EMT inducer
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141632 309.00
Check your price
  • Interferon alpha-2 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517263
  • Size 100µL
Applications
Immunoprecipitation (2 ug), Western Blot (0.1 µg/ml)
Alternate names:
Interferon alpha-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517263 349.00
Check your price
  • CD142 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527278
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Thromboplastin; Coagulation factor III
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527278 349.00
Check your price
  • RBPMS Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA531231
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Heart and RRM expressed sequence
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531231 349.00
Check your price
  • Goat anti-Mouse IgM Cross-Adsorbed Secondary Antibody, DyLight® 350 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate DyLight® 350
  • Catalog # SA510149
  • Size 500ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510149 154.00
Check your price
  • IGFBP1 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA124775
  • Size 100µg
Applications
ELISA (2 µg/ml), Neutralization (10-40 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Placental protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124775 309.00
Check your price
  • Phospho-Cyclin E (Thr62) Polyclonal Antibody for Western Blot, IHC (P), Flow and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517560
  • Size 100µL
Applications
Flow Cytometry (1:50), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
G1/S-specific cyclin-E1; cyclin Es; cyclin Et
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517560 309.00
Check your price
  • NR2C2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # P21960
  • Size 100µL
Applications
Western Blot (Assay Dependent)
Alternate names:
Testicular receptor 4; Orphan nuclear receptor TR4; Orphan nuclear receptor TAK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
P21960 309.00
Check your price