Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  130  140  150  160  170  180  190  200  210  214  215  216  217  218  219  220  221  222  223  224  225  226  227  228  229  230  231  232  233  234  235  236  237  238  239  240  241  242  243  250  260  270  280  290  300  310  320  400  500  600  700  800  900  1000  1100  1200  1622 
6841 - 6870 of 48655 products displayed
  • HIF-2 alpha Polyclonal Antibody, Biotin conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Fish, Hamster, Human, Mouse, Non-human primate, Sheep, Rat
  • Conjugate Biotin
  • Catalog # PA522693
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:200-1:1000)
Alternate names:
Hypoxia-inducible factor 2-alpha; HIF-related factor; HIF-1-alpha-like factor; PAS domain-containing protein 2; Member of PAS protein 2; Class E basic helix-loop-helix protein 73; Basic-helix-loop-helix-PAS protein MOP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522693 375.00
Check your price
  • HIF-2 alpha Monoclonal Antibody for ICC, IHC (P), Flow, IP, ChIP, ELISA and GS
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA116519
  • Size 100 µL
Applications
ChIP assay (Assay-Dependent), ELISA (1:100-1:2000), Flow Cytometry (1:400), Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:150-1:300), Immunoprecipitation (Assay-Dependent), Western Blot (1-2 µg/ml)
Alternate names:
Hypoxia-inducible factor 2-alpha; HIF-related factor; HIF-1-alpha-like factor; PAS domain-containing protein 2; Member of PAS protein 2; Class E basic helix-loop-helix protein 73; Basic-helix-loop-helix-PAS protein MOP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116519 375.00
Check your price
  • HIF-2 alpha Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), Flow, IP, ChIP, ELISA and GS
  • Host Rabbit
  • Target Species Fish, Hamster, Human, Mouse, Non-human primate, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # PA116510
  • Size 100 µL
Applications
ChIP assay (1:10-1:500), ELISA (1:100-1:2000), Flow Cytometry (Assay-Dependent), Gel Shift (Assay-Dependent), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (5 µg/1mg lysate), Western Blot
Alternate names:
Hypoxia-inducible factor 2-alpha; HIF-related factor; HIF-1-alpha-like factor; PAS domain-containing protein 2; Member of PAS protein 2; Class E basic helix-loop-helix protein 73; Basic-helix-loop-helix-PAS protein MOP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116510 375.00
Check your price
  • HIF-2 alpha Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519494
  • Size 100 µg
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin) (1:2500), Western Blot (1 µg/ml)
Alternate names:
Hypoxia-inducible factor 2-alpha; HIF-related factor; HIF-1-alpha-like factor; PAS domain-containing protein 2; Member of PAS protein 2; Class E basic helix-loop-helix protein 73; Basic-helix-loop-helix-PAS protein MOP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519494 349.00
Check your price
  • mTOR Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701483
  • Size 100 µg
Applications
Flow Cytometry (1 ug/ml X 10^6 cells), Immunocytochemistry (1-2 ug/ml), Immunofluorescence (1-2 ug/ml), Western Blot (2-3 ug/ml)
Alternate names:
Rapamycin target protein 1; Rapamycin and FKBP12 target 1; Mechanistic target of rapamycin; Mammalian target of rapamycin; FKBP12-rapamycin complex-associated protein; FK506-binding protein 12-rapamycin complex-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
701483 309.00
Check your price
  • PSMD6 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA530333
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
p42A; Proteasome regulatory particle subunit p44S10; Phosphonoformate immuno-associated protein 4; Breast cancer-associated protein SGA-113M; 26S proteasome regulatory subunit S10; 26S proteasome regulatory subunit RPN7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530333 349.00
Check your price
  • HIP Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517269
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Protein ST13 homolog; Protein FAM10A1; Suppression of tumorigenicity 13 protein; Renal carcinoma antigen NY-REN-33; Putative tumor suppressor ST13; Progesterone receptor-associated p48 protein; Aging-associated protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517269 275.00
Check your price
  • HIP Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513227
  • Size 400 µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Suppression of tumorigenicity 13 protein; Renal carcinoma antigen NY-REN-33; Putative tumor suppressor ST13; Protein FAM10A1; Progesterone receptor-associated p48 protein; Aging-associated protein 2; Protein ST13 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513227 349.00
Check your price
  • Adiponectin Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121170
  • Size 50 µg
Applications
ELISA (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
Gelatin-binding protein; Adipose most abundant gene transcript 1 protein; Adipocyte, C1q and collagen domain-containing protein; Adipocyte complement-related 30 kDa protein; 30 kDa adipocyte complement-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121170 449.00
Check your price
  • Adiponectin Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124411
  • Size 100 µg
Applications
Western Blot (5 µg/ml)
Alternate names:
Gelatin-binding protein; Adipose most abundant gene transcript 1 protein; Adipocyte, C1q and collagen domain-containing protein; Adipocyte complement-related 30 kDa protein; 30 kDa adipocyte complement-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124411 425.00
Check your price
  • ATP6V0A1 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate
  • Catalog # PA525033
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Vacuolar proton translocating ATPase 116 kDa subunit a isoform 1; Vacuolar proton pump subunit 1; Vacuolar adenosine triphosphatase subunit Ac116; Clathrin-coated vesicle/synaptic vesicle proton pump 116 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525033 349.00
Check your price
  • Phospho-BLNK (Tyr84) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA126639
  • Size 50 µL
Applications
Immunohistochemistry (Paraffin) (Assay-dependent), Western Blot (0.1-1 µg/ml)
Alternate names:
Src homology 2 domain-containing leukocyte protein of 65 kDa; Lymphocyte antigen 57; Cytoplasmic adapter protein; B-cell adapter containing a Src homology 2 domain protein; B-cell adapter containing a SH2 domain protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126639 349.00
Check your price
  • Phospho-BLNK (Tyr96) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538357
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Src homology 2 domain-containing leukocyte protein of 65 kDa; Lymphocyte antigen 57; Cytoplasmic adapter protein; B-cell adapter containing a Src homology 2 domain protein; B-cell adapter containing a SH2 domain protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538357 309.00
Check your price
  • BLNK Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515868
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Src homology 2 domain-containing leukocyte protein of 65 kDa; Lymphocyte antigen 57; Cytoplasmic adapter protein; B-cell adapter containing a Src homology 2 domain protein; B-cell adapter containing a SH2 domain protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515868 349.00
Check your price
  • BLNK Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA119377
  • Size 100 µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Src homology 2 domain-containing leukocyte protein of 65 kDa; Lymphocyte antigen 57; Cytoplasmic adapter protein; B-cell adapter containing a Src homology 2 domain protein; B-cell adapter containing a SH2 domain protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA119377 225.00
Check your price
  • BLNK Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131493
  • Size 50 µg
Applications
Western Blot (3-5 µg/ml)
Alternate names:
Src homology 2 domain-containing leukocyte protein of 65 kDa; Lymphocyte antigen 57; Cytoplasmic adapter protein; B-cell adapter containing a Src homology 2 domain protein; B-cell adapter containing a SH2 domain protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131493 399.00
Check your price
  • CD3/CD16/CD56 Monoclonal Antibody, FITC, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC , PE
  • Catalog # MA112207
  • Size 50 Tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain; IgG Fc receptor III-2; FcR-10; Fc-gamma RIII-alpha; CD16a antigen; T-cell surface antigen T3/Leu-4 epsilon chain; IgG Fc receptor III-1; Fc-gamma RIII-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112207 309.00
Check your price
  • CD3/CD16 Monoclonal Antibody, FITC, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC , PE
  • Catalog # MA112085
  • Size 100 Tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain; IgG Fc receptor III-2; FcR-10; Fc-gamma RIII-alpha; CD16a antigen; T-cell surface antigen T3/Leu-4 epsilon chain; IgG Fc receptor III-1; Fc-gamma RIII-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112085 375.00
Check your price
  • CD3/CD16 Monoclonal Antibody, FITC, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC , PE
  • Catalog # MA112086
  • Size 50 Tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain; IgG Fc receptor III-2; FcR-10; Fc-gamma RIII-alpha; CD16a antigen; T-cell surface antigen T3/Leu-4 epsilon chain; IgG Fc receptor III-1; Fc-gamma RIII-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112086 309.00
Check your price
  • CD3e Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate FITC
  • Catalog # A15812
  • Size 50µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
FLJ18683, T3E, TCRE, CD3-epsilon, CD3e antigen, epsilon polypeptide TiT3 complex, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15812 80.50
Check your price
  • CD3e Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate PE
  • Catalog # A15825
  • Size 50µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
FLJ18683, T3E, TCRE, CD3-epsilon, CD3e antigen, epsilon polypeptide TiT3 complex, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15825 89.75
Check your price
  • p19 INK4d Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA527704
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
INK4D, p19, p19-INK4D, CDK inhibitor p19INK4d, cell cycle inhibitor, Nur77 associating protein, cyclin-dependent kinase 4 inhibitor D, cyclin-dependent kinase 4 inhibitor D p19, inhibitor of cyclin-dependent kinase 4d
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527704 349.00
Check your price
  • DNAJB11 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535122
  • Size 400 µL
Applications
Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (1:1000)
Alternate names:
PWP1-interacting protein 4; Human DnaJ protein 9; HEDJ; Endoplasmic reticulum DNA J domain-containing protein 3; ER-associated Hsp40 co-chaperone; ER-associated DNAJ; DnaJ protein homolog 9; APOBEC1-binding protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535122 349.00
Check your price
  • eIF4A3 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530329
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Nuclear matrix protein 265; Eukaryotic translation initiation factor 4A isoform 3; Eukaryotic initiation factor 4A-like NUK-34; DEAD box protein 48; ATP-dependent RNA helicase eIF4A-3; ATP-dependent RNA helicase DDX48
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530329 349.00
Check your price
  • eIF4A3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rat, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA541279
  • Size 100 µl
Applications
Western Blot (1.25 µg/ml)
Alternate names:
Eukaryotic translation initiation factor 4A isoform 3; DEAD box protein 48; ATP-dependent RNA helicase eIF4A-3; ATP-dependent RNA helicase DDX48; Nuclear matrix protein 265; Eukaryotic initiation factor 4A-like NUK-34
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541279 309.00
Check your price
  • IL-12 p70 Monoclonal Antibody for IF, ICC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523715
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml)
Alternate names:
NK cell stimulatory factor chain 1; IL-12 subunit p35; Cytotoxic lymphocyte maturation factor 35 kDa subunit; NK cell stimulatory factor chain 2; IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523715 349.00
Check your price
  • Phospho-FAK2 (Tyr579) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537573
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Related adhesion focal tyrosine kinase; Proline-rich tyrosine kinase 2; Focal adhesion kinase 2; Cell adhesion kinase beta; Calcium-regulated non-receptor proline-rich tyrosine kinase; Calcium-dependent tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537573 309.00
Check your price
  • Phospho-FAK2 (Tyr579, Tyr580) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA126674
  • Size 50 µL
Applications
Western Blot (0.1-1 µg/ml)
Alternate names:
Related adhesion focal tyrosine kinase; Proline-rich tyrosine kinase 2; Focal adhesion kinase 2; Cell adhesion kinase beta; Calcium-regulated non-receptor proline-rich tyrosine kinase; Calcium-dependent tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126674 349.00
Check your price
  • Phospho-FAK2 (Tyr579) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA126673
  • Size 50 µL
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Related adhesion focal tyrosine kinase; Proline-rich tyrosine kinase 2; Focal adhesion kinase 2; Cell adhesion kinase beta; Calcium-regulated non-receptor proline-rich tyrosine kinase; Calcium-dependent tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126673 349.00
Check your price
  • FAK2 Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515407
  • Size 100 µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Related adhesion focal tyrosine kinase; Proline-rich tyrosine kinase 2; Focal adhesion kinase 2; Cell adhesion kinase beta; Calcium-regulated non-receptor proline-rich tyrosine kinase; Calcium-dependent tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515407 349.00
Check your price