Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  31  32  33  34  35  36  37  40  50  60  70  80  90  100  110  120  200  300  400  500  600  700  800  900  1000  1622 
661 - 690 of 48655 products displayed
  • Rabbit anti-Human Kappa Light Chain Secondary Antibody, FITC conjugate for IF
  • Host Rabbit
  • Target Species Human
  • Conjugate FITC
  • Catalog # PA516603
  • Size 2 mL
Applications
Immunofluorescence (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516603 109.00
Check your price
  • Glucocorticoid Receptor Polyclonal Antibody for Western Blot, IF, ICC, IHC, Flow, IP and GS
  • Host Rabbit
  • Target Species Human, Mouse, Reptile, Rat
  • Conjugate Unconjugated
  • Catalog # PA1510A
  • Size 200 µL
Applications
Gel Shift (Assay dependent), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (1:250), Immunoprecipitation (Assay dependent), Western Blot (1:100)
Alternate names:
Nuclear receptor subfamily 3 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1510A 375.00
Check your price
  • CD8 Monoclonal Antibody for IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA180231
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1/20-1/40)
Alternate names:
T-lymphocyte differentiation antigen T8/Leu-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180231 159.00
Check your price
  • MCP-1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA122488
  • Size 50 µg
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (1:200-1:1000), Western Blot (1:2000)
Alternate names:
Small-inducible cytokine A2; Platelet-derived growth factor-inducible protein JE; Monocyte chemotactic protein 1; Monocyte chemoattractant protein 1; Immediate-early serum-responsive protein JE
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA122488 349.00
Check your price
  • HIV1 p24 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17586
  • Size 100 ug
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Human immunodeficiency virus, HIV1gp1, HIV p24
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17586 309.00
Check your price
  • RSV Polyclonal Antibody for IF and ELISA
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA17240
  • Size 1 mL
Applications
ELISA (1:10,000), Immunofluorescence (1:480)
Alternate names:
human respiratory syncytial virus, RSV
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA17240 349.00
Check your price
  • Phospho-CDK1 (Thr14, Tyr15) Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701808
  • Size 100 µg
Applications
Flow Cytometry (2-4 µg x 10^6 cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
p34 protein kinase; Cell division protein kinase 1; Cell division control protein 2 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
701808 309.00
Check your price
  • CD34 Monoclonal Antibody for IHC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA17652
  • Size 100 ug
Applications
Flow Cytometry (1:1000), Immunohistochemistry (Assay dependent)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17652 275.00
Check your price
  • CD44 Monoclonal Antibody for IF, ICC, IHC, Flow, IP and Neu
  • Host Rat
  • Target Species Bovine, Goat, Human, Sheep, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA4400
  • Size 200 µg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Neutralization (Assay Dependent)
Alternate names:
Phagocytic glycoprotein I; Phagocytic glycoprotein 1; Hyaluronate receptor; Hermes antigen; Heparan sulfate proteoglycan; HUTCH-I; GP90 lymphocyte homing/adhesion receptor; Extracellular matrix receptor III; Epican; CDw44
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA4400 349.00
Check your price
  • Phospho-OLIG2 (Ser10, Ser13, Ser14) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA535405
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Protein kinase C-binding protein RACK17; Protein kinase C-binding protein 2; Class E basic helix-loop-helix protein 19; Class B basic helix-loop-helix protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535405 349.00
Check your price
  • Human Interleukin 15 (IL-15) for Western Blot, IP, ELISA and Ctrl
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RIL155
  • Size 10 ug
Applications
Control (Assay Dependent), ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
interleukin-15
Catalog # Size Price (AUD) Your Price (AUD) Qty
RIL155 231.00
Check your price
  • Chlamydia trachomatis MOMP Monoclonal Antibody, FITC conjugate for IF, IHC and ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate FITC
  • Catalog # MA17335
  • Size 100 µg
Applications
ELISA (1:5-1:20), Immunofluorescence (1:5-1:20), Immunohistochemistry (1:5-1:20)
Alternate names:
C. trachomatis major outer membrane protein, C. trachomatis MOMP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17335 375.00
Check your price
  • Human Interleukin 7 (IL-7) for Western Blot, IP, ELISA and Ctrl
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RIL75
  • Size 5 ug
Applications
Control (Assay Dependent), ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Interleukin-7; IL7
Catalog # Size Price (AUD) Your Price (AUD) Qty
RIL75 259.00
Check your price
  • hnRNP A3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA541962
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (4-8 µg/ml), Western Blot (5 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541962 309.00
Check your price
  • Cholera Toxin beta Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA518188
  • Size 100 µg
Applications
ELISA (1:20-1:200)
Alternate names:
beta subunit cholera toxin, CTB, VC1456
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518188 289.00
Check your price
  • IL13RA1 Monoclonal Antibody for IF, ICC, Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA117751
  • Size 100 µg
Applications
ELISA (1:200-1:400), Flow Cytometry (1.2 ug/10^6 cells), Immunocytochemistry (1ug/10^6 cells), Immunofluorescence (1 ug/10^6 cells)
Alternate names:
Cancer/testis antigen 19
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA117751 550.00
Check your price
  • BACE1 Monoclonal Antibody for Western Blot and IF
  • Host Mouse
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA1177
  • Size 100 µg
Applications
Immunofluorescence (1:50-1:200), Western Blot (1:1000-1:2000)
Alternate names:
Membrane-associated aspartic protease 2; Memapsin-2; Beta-site amyloid precursor protein cleaving enzyme 1; Aspartyl protease 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1177 309.00
Check your price
  • SERPINA12 Polyclonal Antibody for Western Blot and IP
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546948
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Visceral adipose-specific serpin; Visceral adipose tissue-derived serine protease inhibitor; OL-64
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546948 375.00
Check your price
  • BCMA Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547794
  • Size 100 µg
Applications
Blocking Assay (Assay-Dependent), Western Blot (0.1 µg/ml)
Alternate names:
B-cell maturation protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547794 349.00
Check your price
  • PAPPA Monoclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # ABS0062402
  • Size 200 µl
Applications
ELISA (1:8,000), Immunohistochemistry (Paraffin) (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Pregnancy-associated plasma protein A; Insulin-like growth factor-dependent IGF-binding protein 4 protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0062402 425.00
Check your price
  • c-Myc Tag Monoclonal Antibody for Western Blot, IF, IHC (P), IP, ELISA and GS
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 132500
  • Size 200 µg
Applications
ELISA (0.1-1.0 ug/ml), Gel Shift (Assay Dependent), Immunofluorescence (2-10 ug/ml), Immunohistochemistry (Assay Dependent), Immunoprecipitation (2-5 ug), Western Blot (1.0 ug/ml)
Alternate names:
Transcription factor p64; Proto-oncogene c-Myc; Class E basic helix-loop-helix protein 39
Catalog # Size Price (AUD) Your Price (AUD) Qty
132500 259.00
Check your price
  • CD45 Monoclonal Antibody for Western Blot, IF, IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513197
  • Size 500 µL
Applications
Flow Cytometry (0.5 ug/test), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:200), Western Blot (1:10-1:100)
Alternate names:
T200; Leukocyte common antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513197 269.00
Check your price
  • POLR2A Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # MA110882
  • Size 200 ug
Applications
Western Blot (Assay dependent)
Alternate names:
DNA-directed RNA polymerase III largest subunit; DNA-directed RNA polymerase II subunit A; RNA-directed RNA polymerase II subunit RPB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110882 375.00
Check your price
  • MRGPRX2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OSR00110W
  • Size 100 µL
Applications
Immunohistochemistry (1:200-1:2000), Western Blot (1:200-1:2000)
Alternate names:
Mas-related G-protein coupled receptor member X2, MRGPRX2, MAS-related GPR, member X2
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00110W 309.00
Check your price
  • beta Amyloid Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511617
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:25-1:50)
Alternate names:
Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI; ABPP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511617 349.00
Check your price
  • HLA-ABC Monoclonal Antibody for IF, IHC (F), Flow, IP, ELISA and FN
  • Host Mouse
  • Target Species Bovine, Human, Non-human primate, Rodent
  • Conjugate Unconjugated
  • Catalog # MA180123
  • Size 500 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (1:50-1:100), Functional Assay (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180123 550.00
Check your price
  • Tau Monoclonal Antibody, Biotin conjugate for Western Blot, IHC and ELISA
  • Host Mouse
  • Target Species Bovine, Human
  • Conjugate Biotin
  • Catalog # MN1000B
  • Size 50 ug
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Paired helical filament-tau; Neurofibrillary tangle protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MN1000B 309.00
Check your price
  • Parathyroid Hormone for Ctrl
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP10937
  • Size 500 µg
Applications
Control (Assay Dependent)
Alternate names:
PTH
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP10937 700.00
Check your price
  • Parainfluenza Virus Type 2 HA Monoclonal Antibody for IF
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA516281
  • Size 2 mg
Applications
Immunofluorescence (Assay Dependent)
Alternate names:
Human parainfluenza virus type 2 , hPIV-2, PIV2 HA, hemagglutinin-Neuraminidase, HPIV2gp7, PIV-2 hemagglutinin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516281 600.00
Check your price
  • PMCA4 ATPase Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Bovine, Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA1914
  • Size 100 µg
Applications
ELISA (1:15,000), Flow Cytometry (2µg per 10^6 cells), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (1:100-1:500), Western Blot (1:1,000)
Alternate names:
Plasma membrane calcium pump isoform 4; Plasma membrane calcium ATPase isoform 4; Matrix-remodeling-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1914 375.00
Check your price