Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  140  150  160  170  180  190  200  210  220  226  227  228  229  230  231  232  233  234  235  236  237  238  239  240  241  242  243  244  245  246  247  248  249  250  251  252  253  254  255  260  270  280  290  300  310  320  330  340  400  500  600  700  800  900  1000  1100  1200  1605 
7201 - 7230 of 48148 products displayed
  • CaMKIV Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517690
  • Size 100µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
CaM kinase-GR; Calspermin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517690 275.00
Check your price
  • CD45R Monoclonal Antibody, Alexa Fluor® 405 conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 405
  • Catalog # RM2626
  • Size 1mL
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
B220, CD45R, Cd45, Ly-5, Lyt-4, T200, Cluster designation 45R
Catalog # Size Price (AUD) Your Price (AUD) Qty
RM2626 332.00
Check your price
  • DR6 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519930
  • Size 100ug
Applications
Western Blot (1:500)
Alternate names:
Death receptor 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519930 349.00
Check your price
  • CEA Monoclonal Antibody for Western Blot, IHC (F), Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA117759
  • Size 100µg
Applications
ELISA (1:200-1:400), Flow Cytometry (1.2 ug/10^6 cells), Immunohistochemistry (Frozen) (1.2 ug/10^6 cells), Western Blot (4 ug/ml)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA117759 550.00
Check your price
  • BNC1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523205
  • Size 100ug
Applications
Western Blot (4-7 µg/ml)
Alternate names:
BNC, BSN1, HsT19447, zinc finger protein basonuclin, zinc finger protein basonuclin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523205 349.00
Check your price
  • PARP2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA14280
  • Size 100µL
Applications
Western Blot (1:250-1:500)
Alternate names:
Poly[ADP-ribose] synthase 2; NAD(+) ADP-ribosyltransferase 2; ADP-ribosyltransferase diphtheria toxin-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14280 375.00
Check your price
  • COL21A1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Dog, Horse, Human, Pig
  • Conjugate Unconjugated
  • Catalog # PA545317
  • Size 100µl
Applications
Western Blot (1 µg/ml )
Alternate names:
collagen, type XXI, alpha 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA545317 309.00
Check your price
  • ERAP2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518948
  • Size 100ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
Leukocyte-derived arginine aminopeptidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518948 349.00
Check your price
  • Mouse Albumin Polyclonal Antibody, HRP conjugate for Western Blot and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # PA130126
  • Size 1mg
Applications
ELISA (1:15,000-1:50,000), Western Blot (1:1,000-1:5,000)
Alternate names:
Alb-1, Alb1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130126 249.00
Check your price
  • CD56 Monoclonal Antibody for IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA119129
  • Size 100µg
Applications
Flow Cytometry (3µg/ml), Immunohistochemistry (Paraffin) (1:50), Immunoprecipitation (Assay Dependent)
Alternate names:
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119129 199.00
Check your price
  • CD4 Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119305
  • Size 100µg
Applications
Flow Cytometry (1 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
T-cell surface antigen T4/Leu-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119305 199.00
Check your price
  • Galectin 4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 425600
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Lactose-binding lectin 4; L-36 lactose-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
425600 309.00
Check your price
  • SLC36A1 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535108
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (1:1000)
Alternate names:
Solute carrier family 36 member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535108 349.00
Check your price
  • CD26 Monoclonal Antibody, Biotin conjugate for Western Blot, IHC (F), Flow and IP
  • Host Mouse
  • Target Species Rat
  • Conjugate Biotin
  • Catalog # MA517548
  • Size 500ug
Applications
Flow Cytometry (2.0 ug/10^6 cells), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
T-cell activation antigen CD26; GP110 glycoprotein; Dipeptidyl peptidase IV; Bile canaliculus domain-specific membrane glycoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517548 309.00
Check your price
  • Phospho-LCK (Ser59) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539760
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539760 309.00
Check your price
  • Phospho-EGFR/ErbB2 (Tyr1173, Tyr1248) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517844
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1; p185erbB2; Tyrosine kinase-type cell surface receptor HER2; Proto-oncogene c-ErbB-2; Proto-oncogene Neu; Metastatic lymph node gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517844 309.00
Check your price
  • Rabbit anti-Human IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate HRP
  • Catalog # A18903
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18903 109.00
Check your price
  • Phospho-PAK4/6/7 (Ser474, Ser602, Ser560) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Guinea pig, Human, Mouse, Pig
  • Conjugate Unconjugated
  • Catalog # PA517636
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
p21-activated kinase 7; p21-activated kinase 5; p21-activated kinase 6; PAK-5; p21-activated kinase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517636 309.00
Check your price
  • Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 350 conjugate for IF, ICC and IHC
  • Host Goat
  • Target Species Rat
  • Conjugate Alexa Fluor® 350
  • Catalog # A21093
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21093 169.00
Check your price
  • Caveolin 1 Monoclonal Antibody for Western Blot, IF and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 036000
  • Size 100µg
Applications
Immunofluorescence (2-4 ug/ml), Immunoprecipitation (2-5 ug/ml), Western Blot (0.5-2.0 ug/ml)
Alternate names:
Phospho-Caveolin-1 pTyr14
Catalog # Size Price (AUD) Your Price (AUD) Qty
036000 365.00
Check your price
  • IGF2R Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # A15744
  • Size 100tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
M6P/IGF2 receptor; Insulin-like growth factor II receptor; Insulin-like growth factor 2 receptor; 300 kDa mannose 6-phosphate receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15744 139.00
Check your price
  • PAK2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515116
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
p58; p21-activated kinase 2; S6/H4 kinase; PAK65; Gamma-PAK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515116 349.00
Check your price
  • beta-1,2 Adaptin Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Bovine, Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA125065
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:200)
Alternate names:
Plasma membrane adaptor HA2/AP2 adaptin beta subunit; Clathrin assembly protein complex 2 beta large chain; Beta-adaptin; Beta-2-adaptin; Adaptor-related protein complex 2 subunit beta; Adaptor protein complex AP-2 subunit beta; AP105B; Golgi adaptor HA1/AP1 adaptin beta subunit; Clathrin assembly protein complex 1 beta large chain; Beta-adaptin 1; Beta-1-adaptin; Adaptor-related protein complex 1 subunit beta-1; Adaptor protein complex AP-1 subunit beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125065 349.00
Check your price
  • ARHGEF12 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539418
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Leukemia-associated RhoGEF
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539418 309.00
Check your price
  • Phospho-HSF1 (Ser303) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537598
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Heat shock transcription factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537598 309.00
Check your price
  • PSMA4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA544542
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Proteasome subunit L; Proteasome component C9; Multicatalytic endopeptidase complex subunit C9; Macropain subunit C9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544542 309.00
Check your price
  • PIH1D1 Monoclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA522090
  • Size 100µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Immunoprecipitation (Assay-Dependent), Western Blot (1-5 µg/ml), Western Blot (Assay-Dependent)
Alternate names:
Nucleolar protein 17 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA522090 309.00
Check your price
  • Ghrelin Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA11046
  • Size 100µg
Applications
Western Blot (1 ug/ml)
Alternate names:
Protein M46; Motilin-related peptide; Growth hormone-releasing peptide; Growth hormone secretagogue
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11046 349.00
Check your price
  • PPP2R1A Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA112384
  • Size 100µg
Applications
Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
PP2A subunit A isoform R1-alpha; PP2A subunit A isoform PR65-alpha; Medium tumor antigen-associated 61 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112384 309.00
Check your price
  • IL-9 Monoclonal Antibody for Western Blot and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523789
  • Size 500µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
T-cell growth factor P40; Cytokine P40
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523789 349.00
Check your price