Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  140  150  160  170  180  190  200  210  220  231  232  233  234  235  236  237  238  239  240  241  242  243  244  245  246  247  248  249  250  251  252  253  254  255  256  257  258  259  260  270  280  290  300  310  320  330  340  400  500  600  700  800  900  1000  1100  1200  1605 
7351 - 7380 of 48138 products displayed
  • mGluR5 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532861
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (5.6 µg/ml)
Alternate names:
GPRC1E, MGLUR5, mGlu5, metabotropic glutamate receptor 5, metabotropic glutamate receptor 5 variant F, metabotropic glutamate receptor 5 variant G, metabotropic glutamate receptor 5 variant H
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532861 349.00
Check your price
  • mGluR5 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532494
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
GPRC1E, MGLUR5, mGlu5, metabotropic glutamate receptor 5, metabotropic glutamate receptor 5 variant F, metabotropic glutamate receptor 5 variant G, metabotropic glutamate receptor 5 variant H
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532494 349.00
Check your price
  • LATS1/LATS2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539741
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Serine/threonine-protein kinase kpm; Large tumor suppressor homolog 2; Kinase phosphorylated during mitosis protein; WARTS protein kinase; Large tumor suppressor homolog 1; Warts-like kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539741 309.00
Check your price
  • LC3A Antibody
  • Host Sheep
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSM00022G
  • Size 500 µg
Applications
Immunohistochemistry (10-50 µg/ml), Western Blot (10-50 µg/ml)
Alternate names:
Microtubule-associated protein 1 light chain 3 alpha; MAP1A/MAP1B light chain 3 A; MAP1 light chain 3-like protein 1; Autophagy-related ubiquitin-like modifier LC3 A; Autophagy-related protein LC3 A
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSM00022G 309.00
Check your price
  • LC3A Antibody
  • Host Sheep
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSA00003W
  • Size 150 µL
Applications
Immunohistochemistry (1:200-1:2000), Western Blot (1:200-1:2000)
Alternate names:
Microtubule-associated protein 1 light chain 3 alpha; MAP1A/MAP1B light chain 3 A; MAP1 light chain 3-like protein 1; Autophagy-related ubiquitin-like modifier LC3 A; Autophagy-related protein LC3 A
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSA00003W 309.00
Check your price
  • LC3A Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSM00029W
  • Size 150 µL
Applications
Immunohistochemistry (1:200-1:2000), Western Blot (1:200-1:2000)
Alternate names:
Microtubule-associated protein 1 light chain 3 alpha; MAP1A/MAP1B light chain 3 A; MAP1 light chain 3-like protein 1; Autophagy-related ubiquitin-like modifier LC3 A; Autophagy-related protein LC3 A
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSM00029W 309.00
Check your price
  • LC3A Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSM00024W
  • Size 150 µL
Applications
Immunohistochemistry (1:100-1:2000), Western Blot (1:100-1:2000)
Alternate names:
Microtubule-associated protein 1 light chain 3 alpha; MAP1A/MAP1B light chain 3 A; MAP1 light chain 3-like protein 1; Autophagy-related ubiquitin-like modifier LC3 A; Autophagy-related protein LC3 A
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSM00024W 309.00
Check your price
  • MAP1LC3C Antibody
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OSM00020W
  • Size 150 µL
Applications
Immunohistochemistry (1:200-1:2000), Western Blot (1:200-1:2000)
Alternate names:
Microtubule-associated protein 1 light chain 3 gamma; MAP1A/MAP1B light chain 3 C; MAP1 light chain 3-like protein 3; Autophagy-related ubiquitin-like modifier LC3 C; Autophagy-related protein LC3 C
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSM00020W 349.00
Check your price
  • MAP1LC3C Antibody
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OSM00021G
  • Size 500 µg
Applications
Immunohistochemistry (10-50 µg/ml), Western Blot (10-50 µg/ml)
Alternate names:
Microtubule-associated protein 1 light chain 3 gamma; MAP1A/MAP1B light chain 3 C; MAP1 light chain 3-like protein 3; Autophagy-related ubiquitin-like modifier LC3 C; Autophagy-related protein LC3 C
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSM00021G 309.00
Check your price
  • MAP1LC3C Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OSM00023W
  • Size 150 µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Microtubule-associated protein 1 light chain 3 gamma; MAP1A/MAP1B light chain 3 C; MAP1 light chain 3-like protein 3; Autophagy-related ubiquitin-like modifier LC3 C; Autophagy-related protein LC3 C
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSM00023W 309.00
Check your price
  • Phospho-p38 MAPK alpha (Thr179, Tyr181) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538416
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538416 309.00
Check your price
  • Phospho-p38 MAPK alpha (Thr180, Tyr182) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFPA0089
  • Size 100 µL
Applications
Western Blot (1:5000)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFPA0089 375.00
Check your price
  • Phospho-p38 MAPK alpha (Thr180, Tyr182) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA112741
  • Size 10 Miniblots
Applications
Dot blot (1:1000), Western Blot (1:1000)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112741 399.00
Check your price
  • Phospho-p38 MAPK pTyr322 Antibody Polyclonal (PA5-35504)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate
  • Catalog # PA535504
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535504 339.00
Check your price
  • p38 MAPK alpha Antibody (Monoclonal, G.876.7)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515204
  • Size 100 µL
Applications
Western Blot (1:2000)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515204 275.00
Check your price
  • p38 MAPK alpha Antibody (Monoclonal, H.477.1)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515069
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515069 275.00
Check your price
  • p38 MAPK alpha Antibody (Monoclonal, J.491.1)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Pig, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # MA515116
  • Size 100 µL
Applications
Flow Cytometry (1:200), Immunocytochemistry (1:25), Immunofluorescence (1:25), Western Blot (1:1000)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515116 275.00
Check your price
  • Phospho-p38 MAPK alpha (Tyr182) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537536
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537536 309.00
Check your price
  • Phospho-p38 MAPK alpha (Thr180) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537535
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537535 309.00
Check your price
  • p38 MAPK alpha Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA514055
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514055 349.00
Check your price
  • p38 MAPK alpha Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517713
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517713 275.00
Check your price
  • p38 MAPK alpha Antibody (Monoclonal, 6A1)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFMA0126
  • Size 100 µL
Applications
Western Blot (4 µg/ml)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0126 309.00
Check your price
  • p38 MAPK alpha Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFPA0160
  • Size 100 µL
Applications
Western Blot (1:2000)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFPA0160 349.00
Check your price
  • p38 MAPK alpha Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA130391
  • Size 1 mL
Applications
Immunohistochemistry (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:200)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130391 425.00
Check your price
  • p38 MAPK alpha Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527831
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527831 349.00
Check your price
  • MINA53 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 409500
  • Size 100 µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Ribosomal oxygenase MINA; Nucleolar protein 52; Mineral dust-induced gene protein; MYC-induced nuclear antigen; Histone lysine demethylase MINA; 60S ribosomal protein L27a histidine hydroxylase
Catalog # Size Price (AUD) Your Price (AUD) Qty
409500 309.00
Check your price
  • NENF Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520577
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Secreted protein of unknown function; Neuron-derived neurotrophic factor; Spinal cord injury-related protein 10; SCIRP10-related protein; Protein GIG47; Cell immortalization-related protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520577 349.00
Check your price
  • NENF Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA545957
  • Size 100 µl
Applications
Western Blot (1 µg/ml )
Alternate names:
Secreted protein of unknown function; Neuron-derived neurotrophic factor; Spinal cord injury-related protein 10; SCIRP10-related protein; Protein GIG47; Cell immortalization-related protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA545957 309.00
Check your price
  • PLEKHF2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531268
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Zinc finger FYVE domain-containing protein 18; Phafin-2; PH and FYVE domain-containing protein 2; Endoplasmic reticulum-associated apoptosis-involved protein containing PH and FYVE domains
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531268 349.00
Check your price
  • Phospho-PP1 alpha (Thr320) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517819
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (1:1000)
Alternate names:
protein phosphatase 1, catalytic subunit, alpha isoform; serine/threonine protein phosphatase PP1-alpha 1 catalytic subunit; serine/threonine-protein phosphatase PP1-alpha catalytic subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517819 309.00
Check your price