Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  150  160  170  180  190  200  210  220  230  234  235  236  237  238  239  240  241  242  243  244  245  246  247  248  249  250  251  252  253  254  255  256  257  258  259  260  261  262  263  270  280  290  300  310  320  330  340  400  500  600  700  800  900  1000  1100  1200  1630 
7441 - 7470 of 48882 products displayed
  • Phospho-JNK1/JNK2 (Thr183, Tyr185) Monoclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Fruit fly, Human, Mouse, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # MA514943
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:50), Immunoprecipitation (1:200), Western Blot (1:1000)
Alternate names:
p54 gamma; c-Jun N-terminal kinase 1; Stress-activated protein kinase JNK1; SAPK gamma; c-Jun N-terminal kinase 2; Stress-activated protein kinase JNK2; Stress-activated protein kinase 1a; JNK-55; Stress-activated protein kinase 1c; JNK-46; p54-alpha; SAPK-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514943 309.00
Check your price
  • Phospho-JNK1/JNK2 (Thr183, Tyr185) Monoclonal Antibody for Western Blot, IF, ICC, Flow, IP and ChIP
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # MA515228
  • Size 200µL
Applications
ChIP assay (1-3 ul), Flow Cytometry (1:200), Immunocytochemistry (1:400), Immunofluorescence (1:400), Immunoprecipitation (1:250), Western Blot (1:2000)
Alternate names:
p54 gamma; c-Jun N-terminal kinase 1; Stress-activated protein kinase JNK1; SAPK gamma; c-Jun N-terminal kinase 2; Stress-activated protein kinase JNK2; Stress-activated protein kinase 1a; JNK-55; Stress-activated protein kinase 1c; JNK-46; p54-alpha; SAPK-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515228 309.00
Check your price
  • Phospho-JNK1/JNK2 (Thr183, Tyr185) Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515175
  • Size 200µL
Applications
Western Blot (1:1000)
Alternate names:
p54 gamma; c-Jun N-terminal kinase 1; Stress-activated protein kinase JNK1; SAPK gamma; c-Jun N-terminal kinase 2; Stress-activated protein kinase JNK2; Stress-activated protein kinase 1a; JNK-55; Stress-activated protein kinase 1c; JNK-46; p54-alpha; SAPK-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515175 309.00
Check your price
  • LAP2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA15828
  • Size 100µL
Applications
Immunocytochemistry (1:250), Immunofluorescence (1:250), Western Blot (1:2500)
Alternate names:
Thymopoietin isoforms alpha/zeta; Thymopoietin isoform beta; Thymopoietin isoforms beta/delta/epsilon/gamma; Thymopoietin-related peptide isoform alpha; Thymopoietin isoform alpha; Thymopoietin-related peptide isoforms beta/gamma; Thymopoietin, isoforms beta/gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA15828 550.00
Check your price
  • Pokemon Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528144
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Zinc finger protein 857A; TTF-I-interacting peptide 21; POZ and Krueppel erythroid myeloid ontogenic factor; Leukemia/lymphoma-related factor; HIV-1 1st-binding protein 1; Factor that binds to inducer of short transcripts protein 1; Factor binding IST protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528144 349.00
Check your price
  • Pokemon Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521087
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Zinc finger protein 857A; TTF-I-interacting peptide 21; POZ and Krueppel erythroid myeloid ontogenic factor; Leukemia/lymphoma-related factor; HIV-1 1st-binding protein 1; Factor that binds to inducer of short transcripts protein 1; Factor binding IST protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521087 349.00
Check your price
  • Pokemon Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131961
  • Size 50µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Zinc finger protein 857A; TTF-I-interacting peptide 21; POZ and Krueppel erythroid myeloid ontogenic factor; Leukemia/lymphoma-related factor; HIV-1 1st-binding protein 1; Factor that binds to inducer of short transcripts protein 1; Factor binding IST protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131961 399.00
Check your price
  • BARD1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524160
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
BARD-1, BRCA1 associated RING domain 1 isoform alfa, BRCA1 associated RING domain 1 isoform beta, BRCA1 associated RING domain 1 isoform delta, BRCA1 associated RING domain 1 isoform epsilon, BRCA1 associated RING domain 1 isoform eta, BRCA1 associated RING dom
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524160 349.00
Check your price
  • CNGB1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Rat
  • Conjugate Unconjugated
  • Catalog # PA1728
  • Size 100µg
Applications
Western Blot (1 ug/ml)
Alternate names:
Glutamic acid-rich protein; Cyclic nucleotide-gated channel beta-1; Cyclic nucleotide-gated cation channel modulatory subunit; Cyclic nucleotide-gated cation channel gamma; Cyclic nucleotide-gated cation channel 4; 240 kDa protein of rod photoreceptor CNG-channel
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1728 349.00
Check your price
  • ERK1/ERK2 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Rabbit
  • Target Species Bovine, Hamster, Human, Mink, Mouse, Non-human primate, Pig, Rat, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # 82380
  • Size 200µL
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
p44-ERK1; Microtubule-associated protein 2 kinase; MNK1; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2; Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1
Catalog # Size Price (AUD) Your Price (AUD) Qty
82380 325.00
Check your price
  • Phospho-JNK1/JNK2 (Thr183, Tyr185) Polyclonal Antibody for Western Blot, ICC and IHC
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA19593
  • Size 30µL
Applications
Immunocytochemistry (1:500-1:2000), Immunohistochemistry (1:500-1:2000), Western Blot (1:2000 -1:4000)
Alternate names:
p54 gamma; c-Jun N-terminal kinase 1; Stress-activated protein kinase JNK1; SAPK gamma; c-Jun N-terminal kinase 2; Stress-activated protein kinase JNK2; Stress-activated protein kinase 1a; JNK-55; Stress-activated protein kinase 1c; JNK-46; p54-alpha; SAPK-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19593 375.00
Check your price
  • Phospho-JNK1/JNK2 (Thr183, Tyr185) Polyclonal Antibody for Western Blot, ICC, IHC and ELISA
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA19594
  • Size 100µL
Applications
ELISA (1:1000-1:4000), Immunocytochemistry (1:500-1:2000), Immunohistochemistry (1:500-1:2000), Western Blot (1:1000-1:4000)
Alternate names:
p54 gamma; c-Jun N-terminal kinase 1; Stress-activated protein kinase JNK1; SAPK gamma; c-Jun N-terminal kinase 2; Stress-activated protein kinase JNK2; Stress-activated protein kinase 1a; JNK-55; Stress-activated protein kinase 1c; JNK-46; p54-alpha; SAPK-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19594 309.00
Check your price
  • ZDHHC17 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518336
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Zinc finger DHHC domain-containing protein 17; Huntingtin-interacting protein 14; Putative NF-kappa-B-activating protein 205; Putative MAPK-activating protein PM11; Huntingtin-interacting protein H; Huntingtin-interacting protein 3; Huntingtin yeast partner H
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518336 349.00
Check your price
  • ZDHHC17 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529942
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Zinc finger DHHC domain-containing protein 17; Huntingtin-interacting protein 14; Putative NF-kappa-B-activating protein 205; Putative MAPK-activating protein PM11; Huntingtin-interacting protein H; Huntingtin-interacting protein 3; Huntingtin yeast partner H
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529942 349.00
Check your price
  • alpha-1 Microglobulin Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527314
  • Size 100µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
A1M, EDC1, HCP, HI30, IATIL, ITI, ITIL, ITILC, UTI, bikunin, complex-forming glycoprotein heterogeneous in charge, growth-inhibiting protein 19, inter-alpha-trypsin inhibitor light chain, protein AMBP, protein HC, trypstatin, uristatin, uronic-acid-rich protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527314 349.00
Check your price
  • ETFB Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519336
  • Size 100ug
Applications
Immunohistochemistry (3-5 ug/ml), Western Blot (1-3 ug/ml)
Alternate names:
ETFB; electron-transfer-flavoprotein, beta polypeptide; FP585; MADD; electron transfer flavoprotein beta subunit; electron transfer flavoprotein beta-subunit; electron transfer flavoprotein, beta polypeptide; electron-transferring-flavoprotein, beta polypeptide
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519336 349.00
Check your price
  • ETFB Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519337
  • Size 100ug
Applications
Immunohistochemistry (3-5 ug/ml), Western Blot (0.01-0.03 ug/ml)
Alternate names:
ETFB; electron-transfer-flavoprotein, beta polypeptide; FP585; MADD; electron transfer flavoprotein beta subunit; electron transfer flavoprotein beta-subunit; electron transfer flavoprotein, beta polypeptide; electron-transferring-flavoprotein, beta polypeptide
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519337 349.00
Check your price
  • MED13 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535924
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Vitamin D3 receptor-interacting protein complex component DRIP250; Thyroid hormone receptor-associated protein complex 240 kDa component; Thyroid hormone receptor-associated protein 1; Mediator complex subunit 13; Activator-recruited cofactor 250 kDa component
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535924 309.00
Check your price
  • RAMP2 Polyclonal Antibody for Western Blot and Flow
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547846
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
CRLR activity-modifying protein 2, calcitonin receptor-like receptor activity modifying protein 2, calcitonin-receptor-like receptor activity-modifying protein 2, receptor (calcitonin) activity modifying protein 2, receptor activity modifying protein 2, recept
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547846 349.00
Check your price
  • CUG-BP1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529923
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
RNA-binding protein BRUNOL-2; Embryo deadenylation element-binding protein homolog; Deadenylation factor CUG-BP; CUG-BP- and ETR-3-like factor 1; CUG triplet repeat RNA-binding protein 1; Bruno-like protein 2; 50 kDa nuclear polyadenylated RNA-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529923 349.00
Check your price
  • CHSY1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530448
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
N-acetylgalactosaminyltransferase 1; N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase 1; Glucuronosyl-N-acetylgalactosaminyl-proteoglycan 4-beta-N-acetylgalactosaminyltransferase 1; Chondroitin synthase 1; Chondroitin glucuronyltransferase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530448 349.00
Check your price
  • CYP1A1/CYP1A2 Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA3036
  • Size 100µL
Applications
Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500)
Alternate names:
Cytochrome P450-P3; Cytochrome P450 4; Cytochrome P(3)450; Cholesterol 25-hydroxylase; CYPIA2; Cytochrome P450-P1; CYPIA1; Cytochrome P450MT2; Cytochrome P450-C; Cytochrome P450-D; Cytochrome P-450d; Cytochrome P-448; Cytochrome P450-P2; Cytochrome P450 form 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3036 349.00
Check your price
  • ETFB Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA522005
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
FP585, MADD, beta-ETF, electron transfer flavoprotein beta subunit, electron transfer flavoprotein beta-subunit, electron transfer flavoprotein subunit beta, electron transfer flavoprotein, beta polypeptide, electron-transferring-flavoprotein, beta polypeptide
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522005 349.00
Check your price
  • hnRNP A2B1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519530
  • Size 100µg
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
DKFZp779B0244, FLJ22720, HNRNPA2, HNRNPB1, HNRPA2, HNRPA2B1, HNRPB1, RNPA2, SNRPB1, heterogeneous nuclear ribonucleoprotein B1; heterogeneous nuclear ribonucleoproteins A2/B1; hnRNP A2 / hnRNP B1; hnRNP A2/B1; nuclear ribonucleoprotein particle A2 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519530 349.00
Check your price
  • Laminin beta-2/gamma-1 Monoclonal Antibody for IF, IHC (F), IHC (P) and IP
  • Host Rat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA514649
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (2 µg/ml), Immunoprecipitation (2 µg/ml)
Alternate names:
S-LAM beta, S-laminin subunit beta, laminin B1s chain, laminin S, laminin subunit beta-2, laminin-11 subunit beta, laminin-14 subunit beta, laminin-15 subunit beta, laminin-3 subunit beta, laminin-4 subunit beta, laminin-7 subunit beta, laminin-9 subunit beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514649 309.00
Check your price
  • LC3B Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA532254
  • Size 100µL
Applications
Flow Cytometry (Assay-Dependent), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B; Microtubule-associated proteins 1A/1B light chain 3B-like
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532254 349.00
Check your price
  • Phospho-ERK1/ERK2 (Thr202, Tyr204, Tyr187) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA126678
  • Size 50µL
Applications
Western Blot (1:1000)
Alternate names:
Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1; p44-ERK1; Microtubule-associated protein 2 kinase; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126678 349.00
Check your price
  • ERK1/ERK2 Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFMA0134
  • Size 100µL
Applications
Western Blot (0.2 µg/ml)
Alternate names:
Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1; p44-ERK1; Microtubule-associated protein 2 kinase; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0134 375.00
Check your price
  • RBCK1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA511949
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Ubiquitin-conjugating enzyme 7-interacting protein 3; UbcM4-interacting protein 28; Heme-oxidized IRP2 ubiquitin ligase 1 homolog; RING finger protein 54; Hepatitis B virus X-associated protein 4; Heme-oxidized IRP2 ubiquitin ligase 1; HBV-associated factor 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511949 349.00
Check your price
  • SEMA4C Polyclonal Antibody for Western Blot and IHC (P)
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547814
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.5-25 µg/ml)
Alternate names:
SEMA4C, SEMACL1, Semaf, SEMAI, FLJ20369, KIAA1739, MGC126382, MGC126383, M-Sema F, M-SEMA-F, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4C, SEMACL1, Semacl1, SEMAF, Semaf, SEMAI, semaphorin-4C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547814 349.00
Check your price