Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  150  160  170  180  190  200  210  220  230  238  239  240  241  242  243  244  245  246  247  248  249  250  251  252  253  254  255  256  257  258  259  260  261  262  263  264  265  266  267  270  280  290  300  310  320  330  340  350  400  500  600  700  800  900  1000  1100  1200  1630 
7561 - 7590 of 48882 products displayed
  • CUG-BP2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA534856
  • Size 100µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
RNA-binding protein BRUNOL-3; Neuroblastoma apoptosis-related RNA-binding protein; ELAV-type RNA-binding protein 3; CUG-BP- and ETR-3-like factor 2; CUG triplet repeat RNA-binding protein 2; Bruno-like protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534856 349.00
Check your price
  • CUG-BP2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA530401
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
RNA-binding protein BRUNOL-3; Neuroblastoma apoptosis-related RNA-binding protein; ELAV-type RNA-binding protein 3; CUG-BP- and ETR-3-like factor 2; CUG triplet repeat RNA-binding protein 2; Bruno-like protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530401 349.00
Check your price
  • Phospho-PKR (Thr258) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539751
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
p68 kinase; Tyrosine-protein kinase EIF2AK2; Protein kinase RNA-activated; P1/eIF-2A protein kinase; Interferon-inducible RNA-dependent protein kinase; Eukaryotic translation initiation factor 2-alpha kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539751 309.00
Check your price
  • PKR Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517785
  • Size 100µL
Applications
Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
p68 kinase; Tyrosine-protein kinase EIF2AK2; Protein kinase RNA-activated; P1/eIF-2A protein kinase; Interferon-inducible RNA-dependent protein kinase; Eukaryotic translation initiation factor 2-alpha kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517785 275.00
Check your price
  • PKR Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517395
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
p68 kinase; Tyrosine-protein kinase EIF2AK2; Protein kinase RNA-activated; P1/eIF-2A protein kinase; Interferon-inducible RNA-dependent protein kinase; Eukaryotic translation initiation factor 2-alpha kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517395 275.00
Check your price
  • ERCC1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518837
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
ERCC1; excision repair cross-complementing rodent repair deficiency, complementation group 1 (includes overlapping antisense sequence); COFS4; UV20; excision repair cross-complementing 1; excision repair protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518837 349.00
Check your price
  • PRDX1 Monoclonal Antibody for Western Blot, Flow and ELISA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # LFMA0073
  • Size 100µL
Applications
ELISA (Assay dependent), Western Blot (1:1000)
Alternate names:
Thioredoxin-dependent peroxide reductase 2; Thioredoxin peroxidase 2; Proliferation-associated gene protein; Natural killer cell-enhancing factor A; Osteoblast-specific factor 3; Macrophage 23 kDa stress protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0073 309.00
Check your price
  • PRDX1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # LFPA0004
  • Size 100µL
Applications
Western Blot (1:1000-1:2000)
Alternate names:
Thioredoxin-dependent peroxide reductase 2; Thioredoxin peroxidase 2; Proliferation-associated gene protein; Natural killer cell-enhancing factor A; Osteoblast-specific factor 3; Macrophage 23 kDa stress protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFPA0004 375.00
Check your price
  • PRDX1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # LFPA0095
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Thioredoxin-dependent peroxide reductase 2; Thioredoxin peroxidase 2; Proliferation-associated gene protein; Natural killer cell-enhancing factor A; Osteoblast-specific factor 3; Macrophage 23 kDa stress protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFPA0095 309.00
Check your price
  • PRDX1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529830
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Thioredoxin-dependent peroxide reductase 2; Thioredoxin peroxidase 2; Proliferation-associated gene protein; Natural killer cell-enhancing factor A; Osteoblast-specific factor 3; Macrophage 23 kDa stress protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529830 349.00
Check your price
  • PSD3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528627
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Pleckstrin homology and SEC7 domain-containing protein 3; Hepatocellular carcinoma-associated antigen 67; Exchange factor for ADP-ribosylation factor guanine nucleotide factor 6; Epididymis tissue protein Li 20mP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528627 349.00
Check your price
  • SLC22A18AS Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531339
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
p27-Beckwith-Wiedemann region 1 B; Solute carrier family 22 member 18 antisense protein; Solute carrier family 22 member 1-like antisense protein; Organic cation transporter-like protein 2 antisense protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531339 349.00
Check your price
  • TAP1 Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA520071
  • Size 100µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
Really interesting new gene 4 protein; Peptide transporter involved in antigen processing 1; Peptide transporter TAP1; Peptide transporter PSF1; Peptide supply factor 1; ATP-binding cassette sub-family B member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA520071 309.00
Check your price
  • TAP1 Polyclonal Antibody for Western Blot, Flow, IP and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131487
  • Size 100µg
Applications
ELISA (Assay-dependent), Flow Cytometry (1:10,000), Immunoprecipitation (1:10,000), Western Blot (1:1000)
Alternate names:
Really interesting new gene 4 protein; Peptide transporter involved in antigen processing 1; Peptide transporter TAP1; Peptide transporter PSF1; Peptide supply factor 1; ATP-binding cassette sub-family B member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131487 550.00
Check your price
  • TAP1 Polyclonal Antibody for Western Blot, IF, ICC, IHC and IP
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA128108
  • Size 50µL
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Assay-dependent), Immunoprecipitation (1:100), Western Blot (1:4000)
Alternate names:
Really interesting new gene 4 protein; Peptide transporter involved in antigen processing 1; Peptide transporter TAP1; Peptide transporter PSF1; Peptide supply factor 1; ATP-binding cassette sub-family B member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128108 399.00
Check your price
  • TAP1 Polyclonal Antibody for Western Blot, Flow, IP and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA186830
  • Size 50µg
Applications
ELISA (1:10000), Flow Cytometry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
Really interesting new gene 4 protein; Peptide transporter involved in antigen processing 1; Peptide transporter TAP1; Peptide transporter PSF1; Peptide supply factor 1; ATP-binding cassette sub-family B member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186830 375.00
Check your price
  • TRIM28 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), ChIP and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA19059
  • Size 100ug
Applications
ChIP assay (Assay Dependent), Immunocytochemistry (5 ug/ml), Immunofluorescence (5 µg/ml), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Tripartite motif-containing protein 28; RING finger protein 96; Nuclear corepressor KAP-1; KRAB-interacting protein 1; KRAB-associated protein 1; E3 SUMO-protein ligase TRIM28; KRIP-1; KRAB-A-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19059 309.00
Check your price
  • USP9X Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA520360
  • Size 100µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
Ubiquitin-specific-processing protease FAF-X; Ubiquitin-specific protease 9, X chromosome; Ubiquitin thioesterase FAF-X; Fat facets protein-related, X-linked; Fat facets in mammals; Deubiquitinating enzyme FAF-X
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA520360 309.00
Check your price
  • DYKDDDDK Tag Monoclonal Antibody, DyLight® 680 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate DyLight® 680
  • Catalog # MA191878D680
  • Size 50µL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (1:500-1:1000)
Alternate names:
FLAG Tag, DYKDDDDK tag, FLAG antibody, ECS epitope tag, DDDDK epitope tag, DDDDK, DYKDDDDK epitope tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, ECS tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191878D680 159.00
Check your price
  • DYKDDDDK Tag Monoclonal Antibody, DyLight® 488 conjugate for IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate DyLight® 488
  • Catalog # MA191878D488
  • Size 50µL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100)
Alternate names:
FLAG Tag, DYKDDDDK tag, FLAG antibody, ECS epitope tag, DDDDK epitope tag, DDDDK, DYKDDDDK epitope tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, ECS tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191878D488 159.00
Check your price
  • Phospho-Aurora B (Thr232) Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA538557
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Serine/threonine-protein kinase aurora-B; Serine/threonine-protein kinase 5; Serine/threonine-protein kinase 12; STK-1; Aurora/IPL1-related kinase 2; Aurora- and IPL1-like midbody-associated protein 1; Aurora 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538557 309.00
Check your price
  • Phospho-Aurora B (Tyr12) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538732
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Serine/threonine-protein kinase aurora-B; Serine/threonine-protein kinase 5; Serine/threonine-protein kinase 12; STK-1; Aurora/IPL1-related kinase 2; Aurora- and IPL1-like midbody-associated protein 1; Aurora 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538732 309.00
Check your price
  • Aurora B Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA517226
  • Size 100µL
Applications
ELISA (Assay Dependent), Western Blot (2 µg/ml)
Alternate names:
Serine/threonine-protein kinase aurora-B; Serine/threonine-protein kinase 5; Serine/threonine-protein kinase 12; STK-1; Aurora/IPL1-related kinase 2; Aurora- and IPL1-like midbody-associated protein 1; Aurora 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517226 349.00
Check your price
  • Aurora B Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Non-human primate, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA186828
  • Size 50µg
Applications
ELISA (1:2000-1:10,000), Western Blot (1:250-1:1000)
Alternate names:
Serine/threonine-protein kinase aurora-B; Serine/threonine-protein kinase 5; Serine/threonine-protein kinase 12; STK-1; Aurora/IPL1-related kinase 2; Aurora- and IPL1-like midbody-associated protein 1; Aurora 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186828 425.00
Check your price
  • Aurora B Polyclonal Antibody for Western Blot, Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517632
  • Size 100µL
Applications
Flow Cytometry (1:50), Immunoprecipitation (1:200), Western Blot (1:1000)
Alternate names:
Serine/threonine-protein kinase aurora-B; Serine/threonine-protein kinase 5; Serine/threonine-protein kinase 12; STK-1; Aurora/IPL1-related kinase 2; Aurora- and IPL1-like midbody-associated protein 1; Aurora 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517632 275.00
Check your price
  • Aurora B Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515321
  • Size 100µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Serine/threonine-protein kinase aurora-B; Serine/threonine-protein kinase 5; Serine/threonine-protein kinase 12; STK-1; Aurora/IPL1-related kinase 2; Aurora- and IPL1-like midbody-associated protein 1; Aurora 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515321 349.00
Check your price
  • Aurora B Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA514075
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Serine/threonine-protein kinase aurora-B; Serine/threonine-protein kinase 5; Serine/threonine-protein kinase 12; STK-1; Aurora/IPL1-related kinase 2; Aurora- and IPL1-like midbody-associated protein 1; Aurora 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514075 349.00
Check your price
  • Aurora B Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA514076
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Serine/threonine-protein kinase aurora-B; Serine/threonine-protein kinase 5; Serine/threonine-protein kinase 12; STK-1; Aurora/IPL1-related kinase 2; Aurora- and IPL1-like midbody-associated protein 1; Aurora 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514076 349.00
Check your price
  • CD44 Oligoclonal Antibody for IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710413
  • Size 100µg
Applications
Flow Cytometry (1 ug/ml X 10^6 cells), Immunocytochemistry (0.5 ug/ml), Immunofluorescence (0.5 ug/ml)
Alternate names:
Phagocytic glycoprotein I; Phagocytic glycoprotein 1; Hyaluronate receptor; Hermes antigen; Heparan sulfate proteoglycan; HUTCH-I; GP90 lymphocyte homing/adhesion receptor; Extracellular matrix receptor III; Epican; CDw44
Catalog # Size Price (AUD) Your Price (AUD) Qty
710413 309.00
Check your price
  • CUG-BP2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA14130
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
RNA-binding protein BRUNOL-3; Neuroblastoma apoptosis-related RNA-binding protein; ELAV-type RNA-binding protein 3; CUG-BP- and ETR-3-like factor 2; CUG triplet repeat RNA-binding protein 2; Bruno-like protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14130 309.00
Check your price