Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  150  160  170  180  190  200  210  220  230  241  242  243  244  245  246  247  248  249  250  251  252  253  254  255  256  257  258  259  260  261  262  263  264  265  266  267  268  269  270  280  290  300  310  320  330  340  350  400  500  600  700  800  900  1000  1100  1200  1630 
7651 - 7680 of 48882 products displayed
  • CRSP9 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528841
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Transcriptional coactivator CRSP33; RNA polymerase transcriptional regulation mediator subunit 7 homolog; Mediator complex subunit 7; Cofactor required for Sp1 transcriptional activation subunit 9; Activator-recruited cofactor 34 kDa component
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528841 349.00
Check your price
  • CRSP9 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA540770
  • Size 100µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin) (1:100-1:200), Western Blot (0.2-1 µg/ml)
Alternate names:
Mediator complex subunit 7; Cofactor required for Sp1 transcriptional activation subunit 9; Transcriptional coactivator CRSP33; RNA polymerase transcriptional regulation mediator subunit 7 homolog; Activator-recruited cofactor 34 kDa component
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540770 309.00
Check your price
  • Opsin Red/Green Polyclonal Antibody for Western Blot
  • Host Chicken
  • Target Species Avian, Cat, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA19517
  • Size 500ug
Applications
Western Blot (1:500-1:2000)
Alternate names:
Medium wavelength-sensitive cone opsin; Green-sensitive opsin; Green cone photoreceptor pigment; Short wavelength-sensitive cone opsin; Blue-sensitive opsin; Blue cone photoreceptor pigment; Red-sensitive opsin; Red cone photoreceptor pigment
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19517 199.00
Check your price
  • Herpes Simplex Virus Type 1 + 2 Envelope Monoclonal Antibody for Western Blot, IF
  • Host Mouse
  • Target Species Virus
  • Conjugate
  • Catalog # MA17265
  • Size 100µg
Applications
Immunofluorescence (1:5), Western Blot (Assay Dependent)
Alternate names:
HSV1, Herpes Simplex I, HSV Type 1, Herpes Simplex 1, HHV, HSV, Herpes Simplex Type 1, HSV2, HSV-2, Human herpes virus 2, HHV-2, Herpes Simplex II, HSV Type 2, Herpes Simplex 2, HSV Type II, HSV 1/2, HSV I/II
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17265 Not Orderable Online
Check your price
  • AKT Pan Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44609G
  • Size 200µL
Applications
Flow Cytometry (3-5 ug x 10^6 cells), Immunocytochemistry (1-3 ug/ml), Immunofluorescence (1-3 ug/ml), Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (0.5-2 ug/ml)
Alternate names:
RAC-PK-beta; Protein kinase B beta; Protein kinase Akt-2; STK-2; RAC-PK-gamma; Protein kinase B gamma; Protein kinase Akt-3; RAC-PK-alpha; Proto-oncogene c-Akt; Protein kinase B alpha; Protein kinase B; Thymoma viral proto-oncogene; AKT1 kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
44609G 309.00
Check your price
  • Bcl-10 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA14129
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
Mammalian CARD-containing adapter molecule E10; Cellular-E10; Cellular homolog of vCARMEN; CED-3/ICH-1 prodomain homologous E10-like regulator; CARD-like apoptotic protein; CARD-containing molecule enhancing NF-kappa-B; B-cell CLL/lymphoma 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14129 309.00
Check your price
  • Phospho-CEBP beta pThr188 + pThr235 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate
  • Catalog # PA535467
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Transcription factor 5; Nuclear factor NF-IL6; Liver-enriched inhibitory protein; Liver activator protein; Silencer factor B; Liver-enriched transcriptional activator; Interleukin-6-dependent-binding protein; C/EBP-related protein 2; AGP/EBP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535467 Not Orderable Online
Check your price
  • CYP2C9 Polyclonal Antibody for Western Blot, IHC (F) and FN
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA184219
  • Size 50µL
Applications
Functional Assay (2-10 µl), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (1:5000-1:10,000)
Alternate names:
S-mephenytoin 4-hydroxylase; Cytochrome P450 PB-1; Cytochrome P450 MP-8; Cytochrome P450 MP-4; Cytochrome P-450MP; Cholesterol 25-hydroxylase; CYPIIC9; (S)-limonene 7-monooxygenase; (S)-limonene 6-monooxygenase; (R)-limonene 6-monooxygenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184219 499.00
Check your price
  • CYP2C9 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515037
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
S-mephenytoin 4-hydroxylase; Cytochrome P450 PB-1; Cytochrome P450 MP-8; Cytochrome P450 MP-4; Cytochrome P-450MP; Cholesterol 25-hydroxylase; CYPIIC9; (S)-limonene 7-monooxygenase; (S)-limonene 6-monooxygenase; (R)-limonene 6-monooxygenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515037 349.00
Check your price
  • CYP2C9 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515046
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
S-mephenytoin 4-hydroxylase; Cytochrome P450 PB-1; Cytochrome P450 MP-8; Cytochrome P450 MP-4; Cytochrome P-450MP; Cholesterol 25-hydroxylase; CYPIIC9; (S)-limonene 7-monooxygenase; (S)-limonene 6-monooxygenase; (R)-limonene 6-monooxygenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515046 349.00
Check your price
  • TIM-1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA141210
  • Size 100µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
T-cell immunoglobulin mucin receptor 1; T cell membrane protein 1; T cell immunoglobulin and mucin domain-containing protein 1; Kidney injury molecule 1; T-cell membrane protein 1; T-cell immunoglobulin and mucin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141210 309.00
Check your price
  • TIM-1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA186790
  • Size 100µg
Applications
Western Blot (1.0-2.0ug/ml)
Alternate names:
T-cell immunoglobulin mucin receptor 1; T cell membrane protein 1; T cell immunoglobulin and mucin domain-containing protein 1; Kidney injury molecule 1; T-cell membrane protein 1; T-cell immunoglobulin and mucin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186790 375.00
Check your price
  • Phospho-AMPK beta-1 (Ser182) Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 700241
  • Size 100µg
Applications
Flow Cytometry (0.5-1 ug/test), Immunocytochemistry (4-6 ug/ml), Immunofluorescence (4-6 ug/ml), Immunohistochemistry (Paraffin) (4-6 ug/ml), Western Blot (2-3 ug/ml)
Alternate names:
5'-AMP-activated protein kinase beta-1 subunit, 5'-AMP-activated protein kinase subunit beta-1, AMP-activated protein kinase beta subunit, AMPK beta -1 chain, AMPK beta 1, AMPK subunit beta-1, AMPKb, protein kinase, AMP-activated, noncatalytic, beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
700241 309.00
Check your price
  • S100A4 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Cat, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA516586
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
S100 calcium-binding protein A4; Protein Mts1; Placental calcium-binding protein; Metastasin; Calvasculin; P9K; Nerve growth factor-induced protein 42A; Placental calcium-binding protein homolog; Protein 18A2; PEL98; Metastatic cell protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516586 375.00
Check your price
  • BRG1 Monoclonal Antibody for Western Blot, IF and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 730011
  • Size 100µg
Applications
Immunofluorescence (1:250), Immunoprecipitation (2 ug), Western Blot (1:2000)
Alternate names:
SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4; SNF2-beta; Protein brahma homolog 1; BRG1-associated factor 190A; ATP-dependent helicase SMARCA4; Protein BRG-1; Mitotic growth and transcription activator
Catalog # Size Price (AUD) Your Price (AUD) Qty
730011 309.00
Check your price
  • BRG1 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 720129
  • Size 100µg
Applications
Flow Cytometry (2-4 µg/million cells), Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4; SNF2-beta; Protein brahma homolog 1; BRG1-associated factor 190A; ATP-dependent helicase SMARCA4; Protein BRG-1; Mitotic growth and transcription activator
Catalog # Size Price (AUD) Your Price (AUD) Qty
720129 304.00
Check your price
  • ADAM33 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528128
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
C20orf153, DJ964F7.1, ADAM 33, a disintegrin and metalloprotease 33, a disintegrin and metalloproteinase domain 33, disintegrin and metalloproteinase domain-containing protein 33, disintegrin and reprolysin metalloproteinase family protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528128 349.00
Check your price
  • PECI Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534898
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Renal carcinoma antigen NY-REN-1; Peroxisomal 3,2-trans-enoyl-CoA isomerase; Hepatocellular carcinoma-associated antigen 88; Dodecenoyl-CoA isomerase; Diazepam-binding inhibitor-related protein 1; Delta(3),delta(2)-enoyl-CoA isomerase; DRS-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534898 349.00
Check your price
  • PECI Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532156
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Renal carcinoma antigen NY-REN-1; Peroxisomal 3,2-trans-enoyl-CoA isomerase; Hepatocellular carcinoma-associated antigen 88; Dodecenoyl-CoA isomerase; Diazepam-binding inhibitor-related protein 1; Delta(3),delta(2)-enoyl-CoA isomerase; DRS-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532156 349.00
Check your price
  • PECI Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532008
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Renal carcinoma antigen NY-REN-1; Peroxisomal 3,2-trans-enoyl-CoA isomerase; Hepatocellular carcinoma-associated antigen 88; Dodecenoyl-CoA isomerase; Diazepam-binding inhibitor-related protein 1; Delta(3),delta(2)-enoyl-CoA isomerase; DRS-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532008 349.00
Check your price
  • eIF3l Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA531647
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Eukaryotic translation initiation factor 3 subunit E-interacting protein; Eukaryotic translation initiation factor 3 subunit 6-interacting protein; PAF67; HSP-66Y; 67 kDa polymerase-associated factor; 66 kDa tyrosine-rich heat shock protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531647 349.00
Check your price
  • XPD Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528916
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Xeroderma pigmentosum group D-complementing protein; TFIIH basal transcription factor complex 80 kDa subunit; DNA repair protein complementing XP-D cells; DNA excision repair protein ERCC-2; CXPD; Basic transcription factor 2 80 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528916 349.00
Check your price
  • XPD Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528236
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Xeroderma pigmentosum group D-complementing protein; TFIIH basal transcription factor complex 80 kDa subunit; DNA repair protein complementing XP-D cells; DNA excision repair protein ERCC-2; CXPD; Basic transcription factor 2 80 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528236 349.00
Check your price
  • XPD Polyclonal Antibody for Western Blot and IF
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519757
  • Size 100µg
Applications
Immunofluorescence (5 µg/ml), Western Blot (2 µg/ml)
Alternate names:
Xeroderma pigmentosum group D-complementing protein; TFIIH basal transcription factor complex 80 kDa subunit; DNA repair protein complementing XP-D cells; DNA excision repair protein ERCC-2; CXPD; Basic transcription factor 2 80 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519757 349.00
Check your price
  • KDM2A Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511177
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
[Histone-H3]-lysine-36 demethylase 1A; JmjC domain-containing histone demethylation protein 1A; F-box/LRR-repeat protein 11; F-box protein Lilina; F-box protein FBL7; F-box and leucine-rich repeat protein 11; CXXC-type zinc finger protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511177 349.00
Check your price
  • KDM2A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511176
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
[Histone-H3]-lysine-36 demethylase 1A; JmjC domain-containing histone demethylation protein 1A; F-box/LRR-repeat protein 11; F-box protein Lilina; F-box protein FBL7; F-box and leucine-rich repeat protein 11; CXXC-type zinc finger protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511176 349.00
Check your price
  • KDM2A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate
  • Catalog # PA511175
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
[Histone-H3]-lysine-36 demethylase 1A; JmjC domain-containing histone demethylation protein 1A; F-box/LRR-repeat protein 11; F-box protein Lilina; F-box protein FBL7; F-box and leucine-rich repeat protein 11; CXXC-type zinc finger protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511175 Not Orderable Online
Check your price
  • KDM2A Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA540421
  • Size 100µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (0.2-1 µg/ml)
Alternate names:
[Histone-H3]-lysine-36 demethylase 1A; JmjC domain-containing histone demethylation protein 1A; F-box/LRR-repeat protein 11; F-box protein Lilina; F-box protein FBL7; F-box and leucine-rich repeat protein 11; CXXC-type zinc finger protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540421 309.00
Check your price
  • PDGFRA Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA110098
  • Size 100ug
Applications
Flow Cytometry (3 ug/ml)
Alternate names:
Platelet-derived growth factor receptor 2; Platelet-derived growth factor alpha receptor; CD140a antigen; CD140 antigen-like family member A; Alpha-type platelet-derived growth factor receptor; Alpha platelet-derived growth factor receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110098 249.00
Check your price
  • PDGFRA Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # MA110097
  • Size 100Tests
Applications
Flow Cytometry (10 ul/10^6 cells)
Alternate names:
Platelet-derived growth factor receptor 2; Platelet-derived growth factor alpha receptor; CD140a antigen; CD140 antigen-like family member A; Alpha-type platelet-derived growth factor receptor; Alpha platelet-derived growth factor receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110097 275.00
Check your price