Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  160  170  180  190  200  210  220  230  240  244  245  246  247  248  249  250  251  252  253  254  255  256  257  258  259  260  261  262  263  264  265  266  267  268  269  270  271  272  273  280  290  300  310  320  330  340  350  400  500  600  700  800  900  1000  1100  1200  1630 
7741 - 7770 of 48882 products displayed
  • 14-3-3 epsilon Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA528937
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:5000-1:20,000)
Alternate names:
14-3-3E, KCIP-1, MDCR, MDS, 14-3-3 epsilon, 14-3-3 protein epsilon, mitochondrial import stimulation factor L subunit, protein kinase C inhibitor protein-1, tyrosine 3/tryptophan 5 -monooxygenase activation protein, epsilon polypeptide
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528937 349.00
Check your price
  • CD8/CD62L Monoclonal Antibody, FITC, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC , PE
  • Catalog # MA112231
  • Size 100Tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
T-lymphocyte differentiation antigen T8/Leu-2; gp90-MEL; TQ1; Lymph node homing receptor; Leukocyte-endothelial cell adhesion molecule 1; Leukocyte surface antigen Leu-8; Leukocyte adhesion molecule 1; CD62 antigen-like family member L
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112231 375.00
Check your price
  • CD8/CD62L Monoclonal Antibody, FITC, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC , PE
  • Catalog # MA112232
  • Size 50Tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
T-lymphocyte differentiation antigen T8/Leu-2; gp90-MEL; TQ1; Lymph node homing receptor; Leukocyte-endothelial cell adhesion molecule 1; Leukocyte surface antigen Leu-8; Leukocyte adhesion molecule 1; CD62 antigen-like family member L
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112232 309.00
Check your price
  • CXCL1 Polyclonal Antibody for Western Blot and ELISA
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA120861
  • Size 50µg
Applications
ELISA (1 µg/ml), Western Blot (1:500)
Alternate names:
Secretory protein N51; Platelet-derived growth factor-inducible protein KC; C-X-C motif chemokine 1; Cytokine-induced neutrophil chemoattractant 1; Neutrophil-activating protein 3; Melanoma growth stimulatory activity; GRO-alpha(1-73)
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120861 464.00
Check your price
  • CYP3A7 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527976
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
CP37, CYPIIIA7, P450-HFLA, aryl hydrocarbon hydroxylase, cytochrome P450 3A7, cytochrome P450, subfamily IIIA, polypeptide 7, cytochrome P450-HFLA, flavoprotein-linked monooxygenase, microsomal monooxygenase, xenobiotic monooxygenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527976 349.00
Check your price
  • XPB Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA123344
  • Size 50µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (1:500)
Alternate names:
Xeroderma pigmentosum group B-complementing protein; TFIIH basal transcription factor complex 89 kDa subunit; DNA repair protein complementing XP-B cells; DNA excision repair protein ERCC-3; Basic transcription factor 2 89 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123344 349.00
Check your price
  • HLA-DR/DP/DQ Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA516983
  • Size 100ug
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
MHC class II antigen DPB1; HLA class II histocompatibility antigen, DP(W4) beta chain; MHC class II antigen DRB1*15; DW2.2/DR2.2; MHC class II antigen DRB1*16; MHC class II antigen DQB1; MHC class II antigen DRB1*3; Clone P2-beta-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516983 319.00
Check your price
  • HLA-DR/DP/DQ Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA180680
  • Size 100Tests
Applications
Flow Cytometry (Neat)
Alternate names:
MHC class II antigen DPB1; HLA class II histocompatibility antigen, DP(W4) beta chain; MHC class II antigen DRB1*15; DW2.2/DR2.2; MHC class II antigen DRB1*16; MHC class II antigen DQB1; MHC class II antigen DRB1*3; Clone P2-beta-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180680 375.00
Check your price
  • HLA-DR/DP/DQ Monoclonal Antibody for IHC (F), IHC (P) and Flow
  • Host Rat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA140077
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
MHC class II antigen DPB1; HLA class II histocompatibility antigen, DP(W4) beta chain; MHC class II antigen DRB1*15; DW2.2/DR2.2; MHC class II antigen DRB1*16; MHC class II antigen DQB1; MHC class II antigen DRB1*3; Clone P2-beta-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140077 375.00
Check your price
  • HLA-DR/DP/DQ Monoclonal Antibody for IHC (P), Flow and FN
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA180678
  • Size 200µg
Applications
Flow Cytometry (1/50-1/200), Functional Assay (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
MHC class II antigen DPB1; HLA class II histocompatibility antigen, DP(W4) beta chain; MHC class II antigen DRB1*15; DW2.2/DR2.2; MHC class II antigen DRB1*16; MHC class II antigen DQB1; MHC class II antigen DRB1*3; Clone P2-beta-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180678 399.00
Check your price
  • HLA-DR/DP/DQ Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA184031
  • Size 200µg
Applications
Flow Cytometry (1/100-1/200)
Alternate names:
MHC class II antigen DPB1; HLA class II histocompatibility antigen, DP(W4) beta chain; MHC class II antigen DRB1*15; DW2.2/DR2.2; MHC class II antigen DRB1*16; MHC class II antigen DQB1; MHC class II antigen DRB1*3; Clone P2-beta-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA184031 399.00
Check your price
  • Galectin 1 Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA530706
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunoprecipitation (1:100-1:500), Western Blot (1:5000-1:20,000)
Alternate names:
S-Lac lectin 1; Lectin galactoside-binding soluble 1; Lactose-binding lectin 1; Galaptin; Beta-galactoside-binding lectin L-14-I; 14 kDa lectin; RL 14.5; Putative MAPK-activating protein PM12; HPL; HBL; 14 kDa laminin-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530706 349.00
Check your price
  • Optineurin Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528249
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
Transcription factor IIIA-interacting protein; Optic neuropathy-inducing protein; NEMO-related protein; Huntingtin-interacting protein L; Huntingtin-interacting protein 7; Huntingtin yeast partner L; FIP-2; E3-14.7K-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528249 349.00
Check your price
  • RPSA Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSR00144W
  • Size 100µL
Applications
Immunohistochemistry (1:300-1:3000), Western Blot (1:300-1:3000)
Alternate names:
Laminin-binding protein precursor p40; Laminin receptor 1; 67 kDa laminin receptor; 37/67 kDa laminin receptor; 37 kDa laminin receptor precursor; NEM/1CHD4; Multidrug resistance-associated protein MGr1-Ag; Colon carcinoma laminin-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00144W 309.00
Check your price
  • RPSA Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSR00145W
  • Size 100µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Laminin-binding protein precursor p40; Laminin receptor 1; 67 kDa laminin receptor; 37/67 kDa laminin receptor; 37 kDa laminin receptor precursor; NEM/1CHD4; Multidrug resistance-associated protein MGr1-Ag; Colon carcinoma laminin-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00145W 349.00
Check your price
  • RPSA Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSR00072W
  • Size 100µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Laminin-binding protein precursor p40; Laminin receptor 1; 67 kDa laminin receptor; 37/67 kDa laminin receptor; 37 kDa laminin receptor precursor; NEM/1CHD4; Multidrug resistance-associated protein MGr1-Ag; Colon carcinoma laminin-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00072W 349.00
Check your price
  • CD263 Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA1956
  • Size 100µg
Applications
Flow Cytometry (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
TRAIL receptor without an intracellular domain; TNF-related apoptosis-inducing ligand receptor 3; Lymphocyte inhibitor of TRAIL; Decoy receptor 1; Decoy TRAIL receptor without death domain; Antagonist decoy receptor for TRAIL/Apo-2L
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1956 349.00
Check your price
  • Herpes Simplex Virus Type 1/2 Polyclonal Antibody, FITC conjugate for IF, ICC and ELISA
  • Host Goat
  • Target Species Virus
  • Conjugate FITC
  • Catalog # PA127209
  • Size 1mL
Applications
ELISA (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50)
Alternate names:
HSV1, Herpes Simplex I, HSV Type 1, Herpes Simplex 1, HHV, HSV, Herpes Simplex Type 1, HSV2, HSV-2, Human herpes virus 2, HHV-2, Herpes Simplex II, HSV Type 2, Herpes Simplex 2, HSV Type II, HSV 1/2, HSV I/II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127209 375.00
Check your price
  • Herpes Simplex Virus Type 1/2 Polyclonal Antibody, HRP conjugate for IF, ICC and ELISA
  • Host Rabbit
  • Target Species Virus
  • Conjugate HRP
  • Catalog # PA127328
  • Size 100µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-dependent)
Alternate names:
HSV1, Herpes Simplex I, HSV Type 1, Herpes Simplex 1, HHV, HSV, Herpes Simplex Type 1, HSV2, HSV-2, Human herpes virus 2, HHV-2, Herpes Simplex II, HSV Type 2, Herpes Simplex 2, HSV Type II, HSV 1/2, HSV I/II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127328 309.00
Check your price
  • ACOT8 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA526152
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Peroxisomal long-chain acyl-CoA thioesterase 1; Peroxisomal acyl-coenzyme A thioester hydrolase 1; Peroxisomal acyl-CoA thioesterase 2; Choloyl-coenzyme A thioesterase; Thioesterase II; PTE-2; HIV-Nef-associated acyl-CoA thioesterase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526152 349.00
Check your price
  • GPIP137 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529514
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
RNA granule protein 105; Membrane component chromosome 11 surface marker 1; GPI-anchored protein p137; GPI-anchored membrane protein 1; Cytoplasmic activation- and proliferation-associated protein 1; Cell cycle-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529514 349.00
Check your price
  • GPIP137 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512179
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
RNA granule protein 105; Membrane component chromosome 11 surface marker 1; GPI-anchored protein p137; GPI-anchored membrane protein 1; Cytoplasmic activation- and proliferation-associated protein 1; Cell cycle-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512179 349.00
Check your price
  • HSPA1B Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528369
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
HSP70-1; HSP70-1A; HSP70I; HSP72; HSPA1; HSP70-1/HSP70-2; HSP70.1/HSP70.2; dnaK-type molecular chaperone HSP70-1; heat shock 70 kDa protein 1/2; heat shock 70 kDa protein 1A/1B; heat shock 70kD protein 1A; heat shock-induced protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528369 349.00
Check your price
  • HSPA1B Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529605
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
HSP70-1; HSP70-1A; HSP70I; HSP72; HSPA1; HSP70-1/HSP70-2; HSP70.1/HSP70.2; dnaK-type molecular chaperone HSP70-1; heat shock 70 kDa protein 1/2; heat shock 70 kDa protein 1A/1B; heat shock 70kD protein 1A; heat shock-induced protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529605 349.00
Check your price
  • PCQAP Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519081
  • Size 100µg
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Trinucleotide repeat-containing gene 7 protein; TPA-inducible gene 1 protein; Positive cofactor 2 glutamine/Q-rich-associated protein; Mediator complex subunit 15; CTG repeat protein 7a; Activator-recruited cofactor 105 kDa component
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519081 349.00
Check your price
  • PLRG1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA517990
  • Size 100ug
Applications
Western Blot (0.5-2 ug/ml)
Alternate names:
PLRG1; pleiotropic regulator 1 (PRL1 homolog, Arabidopsis); PRL1; MGC110980; pleiotropic regulator 1 (PRL1, Arabidopsis homolog); pleiotropic regulator 1 (PRL1homolog, Arabidopsis); pleiotropic regulator 1 (PRL1, Arabidopsis homolog)
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517990 349.00
Check your price
  • COX2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517614
  • Size 100µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Assay dependent), Western Blot (1:1000)
Alternate names:
Prostaglandin-endoperoxide synthase 2; Prostaglandin H2 synthase 2; PHS II; Cyclooxygenase-2; TIS10 protein; PES-2; Macrophage activation-associated marker protein P71/73; Gripghs; Glucocorticoid-regulated inflammatory cyclooxygenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517614 275.00
Check your price
  • COX2 Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA516817
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100-200), Immunoprecipitation (Assay Dependent), Western Blot (2-4 µg/ml)
Alternate names:
Prostaglandin-endoperoxide synthase 2; Prostaglandin H2 synthase 2; PHS II; Cyclooxygenase-2; TIS10 protein; PES-2; Macrophage activation-associated marker protein P71/73; Gripghs; Glucocorticoid-regulated inflammatory cyclooxygenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516817 309.00
Check your price
  • HCC1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA531103
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Splicing factor HCC1; RNA-binding region-containing protein 2; RNA-binding motif protein 39; Hepatocellular carcinoma protein 1; CAPER alpha; Transcription coactivator CAPER; Coactivator of activating protein 1 and estrogen receptors
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531103 349.00
Check your price
  • TGIF2 Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA522442
  • Size 100µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
5'-TG-3' interacting factor 2, TGF(beta)-induced transcription factor 2, TGF-beta-induced transcription factor 2, TGFB-induced factor 2 (TALE family homeobox), homeobox protein TGIF2, transcription growth factor-beta-induced factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA522442 309.00
Check your price