Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  170  180  190  200  210  220  230  240  250  261  262  263  264  265  266  267  268  269  270  271  272  273  274  275  276  277  278  279  280  281  282  283  284  285  286  287  288  289  290  300  310  320  330  340  350  360  370  400  500  600  700  800  900  1000  1100  1200  1630 
8251 - 8280 of 48882 products displayed
  • CPN1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA543747
  • Size 100µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (0.2-1 µg/ml)
Alternate names:
Serum carboxypeptidase N; Plasma carboxypeptidase B; Lysine carboxypeptidase; Kininase-1; Carboxypeptidase N small subunit; Carboxypeptidase N polypeptide 1; Arginine carboxypeptidase; Anaphylatoxin inactivator
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543747 309.00
Check your price
  • CPN1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA514222
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
Serum carboxypeptidase N; Plasma carboxypeptidase B; Lysine carboxypeptidase; Kininase-1; Carboxypeptidase N small subunit; Carboxypeptidase N polypeptide 1; Arginine carboxypeptidase; Anaphylatoxin inactivator
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514222 349.00
Check your price
  • CYP4F2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Pig
  • Conjugate Unconjugated
  • Catalog # PA527769
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Leukotriene-B(4) omega-hydroxylase 1; Leukotriene-B(4) 20-monooxygenase 1; Cytochrome P450-LTB-omega; Cytochrome P450 4F2; CYPIVF2; Arachidonic acid omega-hydroxylase; 20-hydroxyeicosatetraenoic acid synthase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527769 349.00
Check your price
  • DDR2 Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515356
  • Size 100µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Tyrosine-protein kinase TYRO10; Receptor protein-tyrosine kinase TKT; Neurotrophic tyrosine kinase, receptor-related 3; Discoidin domain-containing receptor tyrosine kinase 2; CD167 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515356 349.00
Check your price
  • DDR2 Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131993
  • Size 100µg
Applications
ELISA (1:1000), Western Blot (1:100-1:500)
Alternate names:
Tyrosine-protein kinase TYRO10; Receptor protein-tyrosine kinase TKT; Neurotrophic tyrosine kinase, receptor-related 3; Discoidin domain-containing receptor tyrosine kinase 2; CD167 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131993 449.00
Check your price
  • DDR2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527752
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Tyrosine-protein kinase TYRO10; Receptor protein-tyrosine kinase TKT; Neurotrophic tyrosine kinase, receptor-related 3; Discoidin domain-containing receptor tyrosine kinase 2; CD167 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527752 349.00
Check your price
  • DDR2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527730
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Tyrosine-protein kinase TYRO10; Receptor protein-tyrosine kinase TKT; Neurotrophic tyrosine kinase, receptor-related 3; Discoidin domain-containing receptor tyrosine kinase 2; CD167 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527730 349.00
Check your price
  • DDR2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA514751
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
Tyrosine-protein kinase TYRO10; Receptor protein-tyrosine kinase TKT; Neurotrophic tyrosine kinase, receptor-related 3; Discoidin domain-containing receptor tyrosine kinase 2; CD167 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514751 349.00
Check your price
  • H6PD Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527439
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
G6PDH, GDH, 6-phosphogluconolactonase, G6PD, H form, GDH/6PGL endoplasmic bifunctional protein, glucose 1- dehydrogenase, glucose dehydrogenase, glucose dehyrogenase, glucose-6-phosphate dehydrogenase, salivary
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527439 349.00
Check your price
  • H6PD Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA524960
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
G6PDH, GDH, 6-phosphogluconolactonase, G6PD, H form, GDH/6PGL endoplasmic bifunctional protein, glucose 1- dehydrogenase, glucose dehydrogenase, glucose dehyrogenase, glucose-6-phosphate dehydrogenase, salivary
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524960 349.00
Check your price
  • RALY Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA530929
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
hnRNP associated with lethal yellow protein; Maternally-expressed hnRNP C-related protein; hnRNP associated with lethal yellow protein homolog; Heterogeneous nuclear ribonucleoprotein C-like 2; Autoantigen p542
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530929 349.00
Check your price
  • RALY Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Goat, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA540822
  • Size 100µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (0.2-1 µg/ml)
Alternate names:
hnRNP associated with lethal yellow protein; Maternally-expressed hnRNP C-related protein; hnRNP associated with lethal yellow protein homolog; Heterogeneous nuclear ribonucleoprotein C-like 2; Autoantigen p542
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540822 309.00
Check your price
  • RALY Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA540651
  • Size 100µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (1.25 µg/ml)
Alternate names:
hnRNP associated with lethal yellow protein; Maternally-expressed hnRNP C-related protein; hnRNP associated with lethal yellow protein homolog; Heterogeneous nuclear ribonucleoprotein C-like 2; Autoantigen p542
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540651 309.00
Check your price
  • RUVBL2 Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA534608
  • Size 100µg
Applications
Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin) (1:400), Western Blot (1 µg/ml)
Alternate names:
zReptin; Reptin; p47 protein; TIP60-associated protein 54-beta; TIP49b; Repressing pontin 52; INO80 complex subunit J; 51 kDa erythrocyte cytosolic protein; 48 kDa TATA box-binding protein-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534608 349.00
Check your price
  • S100A8 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA517622
  • Size 100ug
Applications
Flow Cytometry (0.5 ug/10^6 cells)
Alternate names:
Urinary stone protein band A; S100 calcium-binding protein A8; Migration inhibitory factor-related protein 8; Leukocyte L1 complex light chain; Cystic fibrosis antigen; Calprotectin L1L subunit; Calgranulin-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517622 159.00
Check your price
  • S100A8 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517621
  • Size 200ug
Applications
Flow Cytometry (0.5 ug/10^6 cells)
Alternate names:
Urinary stone protein band A; S100 calcium-binding protein A8; Migration inhibitory factor-related protein 8; Leukocyte L1 complex light chain; Cystic fibrosis antigen; Calprotectin L1L subunit; Calgranulin-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517621 225.00
Check your price
  • NHERF2 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518083
  • Size 100µL
Applications
Immunoprecipitation (Assay-dependent), Western Blot (1:100)
Alternate names:
Tyrosine kinase activator protein 1; Solute carrier family 9 isoform A3 regulatory factor 2; Sodium-hydrogen exchanger regulatory factor 2; SRY-interacting protein 1; NHE3 kinase A regulatory protein E3KARP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518083 399.00
Check your price
  • NHERF2 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518436
  • Size 100ug
Applications
Immunohistochemistry (1-2µg/ml), Western Blot (0.3-1 ug/ml)
Alternate names:
Tyrosine kinase activator protein 1; Solute carrier family 9 isoform A3 regulatory factor 2; Sodium-hydrogen exchanger regulatory factor 2; SRY-interacting protein 1; NHE3 kinase A regulatory protein E3KARP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518436 349.00
Check your price
  • Aurora B Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 365200
  • Size 100µg
Applications
Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (1µg/mL), Immunofluorescence (1µg/mL), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Serine/threonine-protein kinase aurora-B; Serine/threonine-protein kinase 5; Serine/threonine-protein kinase 12; STK-1; Aurora/IPL1-related kinase 2; Aurora- and IPL1-like midbody-associated protein 1; Aurora 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
365200 304.00
Check your price
  • BAZ1A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA540610
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
hWALp1; Williams syndrome transcription factor-related chromatin-remodeling factor 180; CHRAC subunit ACF1; ATP-utilizing chromatin assembly and remodeling factor 1; ATP-dependent chromatin-remodeling protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540610 309.00
Check your price
  • IRGM Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520508
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
LPS-stimulated RAW 264.7 macrophage protein 47; Interferon-inducible protein 1; Interferon-inducible GTPase 3; LPS-stimulated RAW 264.7 macrophage protein 47 homolog; Immunity-related GTPase family M protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520508 349.00
Check your price
  • IRGM Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520507
  • Size 100ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
LPS-stimulated RAW 264.7 macrophage protein 47; Interferon-inducible protein 1; Interferon-inducible GTPase 3; LPS-stimulated RAW 264.7 macrophage protein 47 homolog; Immunity-related GTPase family M protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520507 349.00
Check your price
  • LXR beta Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520886
  • Size 100µg
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Ubiquitously-expressed nuclear receptor; Retinoid X receptor-interacting protein No.15; Nuclear receptor subfamily 1 group H member 2; Liver X receptor beta; Nuclear receptor NER; Orphan nuclear receptor OR-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520886 349.00
Check your price
  • Olfr156 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # OSR00044W
  • Size 100µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Olfactory receptor 156, MOR262-3, MOR262-5, MOR262-7, MOR262-11, MOR262-12, Olfactory receptor MOR262-6, Olfactory receptor 155, Olfr156, Olfr157, Olf, MOR262-6, MOR262- 3, MOR262- 5, MOR262- 7, MOR262- 11, MOR262- 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00044W 349.00
Check your price
  • PDZK1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530120
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Sodium-hydrogen exchanger regulatory factor 3; PDZ domain-containing protein 1; Na/Pi cotransporter C-terminal-associated protein 1; Na(+)/H(+) exchanger regulatory factor 3; CFTR-associated protein of 70 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530120 349.00
Check your price
  • Phospho-FAK2 (Tyr402) Oligoclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710087
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (4-5 ug/ml)
Alternate names:
Related adhesion focal tyrosine kinase; Proline-rich tyrosine kinase 2; Focal adhesion kinase 2; Cell adhesion kinase beta; Calcium-regulated non-receptor proline-rich tyrosine kinase; Calcium-dependent tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
710087 309.00
Check your price
  • RRS1 Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524677
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
homolog of yeast ribosome biogenesis regulator, homolog of yeast ribosome biogenesis regulatory protein RRS1, ribosome biogenesis regulatory protein RRS1 homolog, ribosome biogenesis regulatory protein homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524677 349.00
Check your price
  • SERPINH1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527832
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Rheumatoid arthritis-related antigen RA-A47; Collagen-binding protein; Cell proliferation-inducing gene 14 protein; Arsenic-transactivated protein 3; 47 kDa heat shock protein; Serine protease inhibitor J6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527832 349.00
Check your price
  • NSD2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA540725
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Wolf-Hirschhorn syndrome candidate 1 protein homolog; Nuclear SET domain-containing protein 2; Multiple myeloma SET domain-containing protein; Wolf-Hirschhorn syndrome candidate 1 protein; Protein trithorax-5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540725 309.00
Check your price
  • DYKDDDDK Tag Monoclonal Antibody for Western Blot and ELISA
  • Host Rat
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA11421MG
  • Size 1mg
Applications
ELISA (100ng/ml), Western Blot (1ug/ml)
Alternate names:
FLAG Tag, DYKDDDDK tag, FLAG antibody, ECS epitope tag, DDDDK epitope tag, DDDDK, DYKDDDDK epitope tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, ECS tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA11421MG 900.00
Check your price