Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  31  32  33  34  35  36  37  38  39  40  41  42  50  60  70  80  90  100  110  120  200  300  400  500  600  700  800  900  1000  1630 
811 - 840 of 48882 products displayed
  • CD49f Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517088
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
VLA-6; CD49 antigen-like family member F
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517088 275.00
Check your price
  • CD4 Monoclonal Antibody for Flow and IP
  • Host Mouse
  • Target Species Horse
  • Conjugate Unconjugated
  • Catalog # MA181196
  • Size 2mL
Applications
Flow Cytometry (Neat), Immunoprecipitation (Assay dependent)
Alternate names:
CD4 antigen p55, CD4 receptor, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181196 375.00
Check your price
  • CXCR4 Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519856
  • Size 100ug
Applications
Flow Cytometry (0.5 µg/ml), Immunocytochemistry (20 µg/ml), Immunofluorescence (20 µg/ml), Immunoprecipitation (10 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Stromal cell-derived factor 1 receptor; NPYRL; Lipopolysaccharide-associated protein 3; Leukocyte-derived seven transmembrane domain receptor; LCR1; HM89; Fusin; FB22; Pre-B-cell-derived chemokine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519856 349.00
Check your price
  • MCP-1 Polyclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513658
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Small-inducible cytokine A2; Monocyte secretory protein JE; Monocyte chemotactic protein 1; Monocyte chemotactic and activating factor; Monocyte chemoattractant protein 1; HC11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513658 349.00
Check your price
  • FMN2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Mouse, Human
  • Conjugate Unconjugated
  • Catalog # PA523238
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
RP11-90L13.1, formin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523238 349.00
Check your price
  • Ovalbumin Polyclonal Antibody, HRP conjugate for Western Blot and ELISA
  • Host Rabbit
  • Target Species Chicken
  • Conjugate HRP
  • Catalog # PA1196HRP
  • Size 100µg
Applications
ELISA (1:1000-1:5000), Western Blot (1:1000-1:5000)
Alternate names:
Plakalbumin; Egg albumin; Allergen Gal d II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1196HRP 225.00
Check your price
  • Phospho-CXCR4 (Ser339) Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538461
  • Size 100µg
Applications
Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Stromal cell-derived factor 1 receptor; NPYRL; Lipopolysaccharide-associated protein 3; Leukocyte-derived seven transmembrane domain receptor; LCR1; HM89; Fusin; FB22; Pre-B-cell-derived chemokine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538461 309.00
Check your price
  • beta Actin Monoclonal Antibody, Alexa Fluor® 647 conjugate for Western Blot
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Alexa Fluor® 647
  • Catalog # MA1140A647
  • Size 50µl
Applications
Western Blot (Assay-Dependent)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1140A647 225.00
Check your price
  • ZKSCAN4 Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523296
  • Size 100µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
Zinc finger protein 427; Zinc finger protein 307; P373c6.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523296 309.00
Check your price
  • CXCL12 Polyclonal Antibody for Western Blot, IHC and IP
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA129029
  • Size 100µg
Applications
Immunohistochemistry (1:100-1:500), Immunoprecipitation (1:300-1:500), Western Blot (1:1000)
Alternate names:
Thymic lymphoma cell-stimulating factor; Pre-B cell growth-stimulating factor; C-X-C motif chemokine 12; 12-O-tetradecanoylphorbol 13-acetate repressed protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129029 349.00
Check your price
  • CD25 Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517815
  • Size 100ug
Applications
Flow Cytometry (0.2 ug/10^6 cells)
Alternate names:
p55
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517815 119.00
Check your price
  • Rabbit anti-Human IgG Fc Secondary Antibody, HRP conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate HRP
  • Catalog # 31423
  • Size 1.5mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:100,000), Immunofluorescence (1:5,000-1:100,000), Immunohistochemistry (1:5,000-1:100,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:200,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31423 199.00
Check your price
  • CD38 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA119523
  • Size 100µg
Applications
Flow Cytometry (1:500)
Alternate names:
T10; Cyclic ADP-ribose hydrolase 1; ADP-ribosyl cyclase 1; 2'-phospho-cyclic-ADP-ribose transferase; 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase; 2'-phospho-ADP-ribosyl cyclase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119523 225.00
Check your price
  • Cytokeratin 14 Polyclonal Antibody for Western Blot, IF, ICC, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA528002
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Frozen) (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Keratin-14; Cytokeratin-14; Type I keratin Ka14
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528002 349.00
Check your price
  • CXCL5 Oligoclonal Antibody for Western Blot, IHC (P), Flow and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710010
  • Size 100µg
Applications
Flow Cytometry (3-5 µg/million cells), Immunohistochemistry (Paraffin) (1:10-1:50), Immunoprecipitation (5 µg), Western Blot (2-5 µg/ml)
Alternate names:
Small-inducible cytokine B5; Neutrophil-activating peptide ENA-78; Epithelial-derived neutrophil-activating protein 78; ENA-78(1-78)
Catalog # Size Price (AUD) Your Price (AUD) Qty
710010 309.00
Check your price
  • RUNX2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Chicken, Horse, Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA141519
  • Size 100µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
SL3/AKV core-binding factor alpha A subunit; SL3-3 enhancer factor 1 alpha A subunit; Polyomavirus enhancer-binding protein 2 alpha A subunit; Osteoblast-specific transcription factor 2; Oncogene AML-3; Core-binding factor subunit alpha-1; Acute myeloid leukemia 3 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141519 375.00
Check your price
  • Calretinin Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA139562
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
29 kDa calbindin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA139562 425.00
Check your price
  • Treponema pallidum Polyclonal Antibody for IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # PA17257
  • Size 1mL
Applications
Immunohistochemistry (Paraffin, Frozen) (1:3,000)
Alternate names:
T. pallidum, Syphillis
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA17257 309.00
Check your price
  • Entamoeba histolytica Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Protozoa
  • Conjugate Unconjugated
  • Catalog # MA110657
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
E. histolytica
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110657 349.00
Check your price
  • TRIB1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514801
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
SKIP1; G-protein-coupled receptor-induced gene 2 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514801 349.00
Check your price
  • PP2CA/PP2CB Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA112395
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Protein phosphatase 2C isoform beta; Protein phosphatase IA; Protein phosphatase 2C isoform alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112395 309.00
Check your price
  • Ferritin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191849
  • Size 500µg
Applications
ELISA (Assay Dependent)
Alternate names:
MGC71996
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191849 325.00
Check your price
  • Asialo GM1 Polyclonal Antibody for ELISA
  • Host Rabbit
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # PA185210
  • Size 1mL
Applications
ELISA (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185210 425.00
Check your price
  • Lamin B1 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), Flow and ELISA
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Sheep, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA106103
  • Size 100µL
Applications
ELISA (Assay Dependent), Flow Cytometry (1:100-1:200), Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
ADLD, LMN, LMN2, LMNB, MGC111419
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106103 375.00
Check your price
  • CX3CL1 Polyclonal Antibody for Western Blot, IHC (F), ELISA and Neu
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA547728
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunohistochemistry (Frozen) (5-15 µg/ml), Neutralization (0.3-1.2 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Small-inducible cytokine D1; Neurotactin; CX3C membrane-anchored chemokine; C-X3-C motif chemokine 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547728 349.00
Check your price
  • AFP Oligoclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710486
  • Size 100µg
Applications
Immunocytochemistry (1-2 ug/ml), Immunofluorescence (1-2 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Alpha-fetoglobulin; Alpha-1-fetoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
710486 309.00
Check your price
  • SHH Monoclonal Antibody for Western Blot, IHC, ELISA and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523883
  • Size 100µg
Applications
ELISA (0.5-2 µg/ml (Detection), 2-8 µg/ml (Capture)), Immunohistochemistry (8-25 µg/ml), Neutralization (5-15 µg/ml), Western Blot (2 µg/ml)
Alternate names:
HHG-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523883 275.00
Check your price
  • PRDX6 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFPA0005
  • Size 100µL
Applications
Western Blot (1:2000)
Alternate names:
Red blood cells page spot 12; Non-selenium glutathione peroxidase; Liver 2D page spot 40; Antioxidant protein 2; Acidic calcium-independent phospholipase A2; 24 kDa protein; 1-Cys peroxiredoxin
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFPA0005 375.00
Check your price
  • Phospho-PLM (Ser68) Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA511124
  • Size 400µL
Applications
Western Blot (1:100-1:500)
Alternate names:
FXYD domain-containing ion transport regulator 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511124 349.00
Check your price
  • SAM68 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519546
  • Size 100µg
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Western Blot (1 µg/ml)
Alternate names:
p68; p21 Ras GTPase-activating protein-associated p62; Src-associated in mitosis 68 kDa protein; GAP-associated tyrosine phosphoprotein p62
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519546 349.00
Check your price