Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  19  20  21  22  23  24  25  26  27  28  29  30  31  32  33  34  35  36  37  38  39  40  41  42  43  44  45  46  47  48  50  60  70  80  90  100  110  120  130  200  300  400  500  600  700  800  900  1000  1611 
991 - 1020 of 48310 products displayed
  • Goat anti-Human IgG Fc Secondary Antibody, HRP conjugate
  • Host Goat
  • Target Species Human
  • Conjugate HRP
  • Catalog # H10007
  • Size 2 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
H10007 164.00
Check your price
  • FER Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522992
  • Size 100 µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Western Blot (0.5 µg/ml)
Alternate names:
p94-Fer; Tyrosine kinase 3; Proto-oncogene c-Fer; Fujinami poultry sarcoma/Feline sarcoma-related protein Fer; Feline encephalitis virus-related kinase FER
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522992 349.00
Check your price
  • Goat anti-Human IgG (H+L) Cross-Adsorbed Secondary Antibody for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 31119
  • Size 1.5 mg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31119 104.00
Check your price
  • Phospho-ERK1/ERK2 (Tyr204) Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Fruit fly, Human, Mink, Mouse, Non-human primate, Pig, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA515174
  • Size 200 µL
Applications
Flow Cytometry (1:200), Immunocytochemistry (1:200), Immunofluorescence (1:200), Western Blot (1:1000)
Alternate names:
p44-ERK1; Microtubule-associated protein 2 kinase; MNK1; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2; Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1; Protein rolled; Extracellular-regulated kinase A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515174 309.00
Check your price
  • Claudin 5 Monoclonal Antibody, Alexa Fluor® 488 conjugate for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Alexa Fluor® 488
  • Catalog # 352588
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Lung-specific membrane protein; Brain endothelial cell clone 1 protein; Transmembrane protein deleted in VCFS
Catalog # Size Price (AUD) Your Price (AUD) Qty
352588 328.00
Check your price
  • Factor X Monoclonal Antibody for IA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA143013
  • Size 100 ug
Applications
Inhibition Assays (Assay Dependent.)
Alternate names:
Stuart-Prower factor; Stuart factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA143013 249.00
Check your price
  • Bcl-10 Monoclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA514878
  • Size 100 µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Mammalian CARD-containing adapter molecule E10; Cellular-E10; Cellular homolog of vCARMEN; CED-3/ICH-1 prodomain homologous E10-like regulator; CARD-like apoptotic protein; CARD-containing molecule enhancing NF-kappa-B; B-cell CLL/lymphoma 10; R-RCD1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514878 275.00
Check your price
  • CD45 Monoclonal Antibody, TRI-COLOR® conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate TRI-COLOR®
  • Catalog # MHCD45065
  • Size 2.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
T200; Leukocyte common antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD45065 1,248.00
Check your price
  • Adenylate Cyclase 9 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131194
  • Size 100 µg
Applications
Western Blot (1:500)
Alternate names:
Adenylyl cyclase type 10; Adenylyl cyclase 9; Adenylate cyclase type IX; ATP pyrophosphate-lyase 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131194 449.00
Check your price
  • NLRP3 Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA523919
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Western Blot (2 µg/ml)
Alternate names:
PYRIN-containing APAF1-like protein 1; Cryopyrin; Cold-induced autoinflammatory syndrome 1 protein; Caterpiller protein 1.1; Angiotensin/vasopressin receptor AII/AVP-like; Mast cell maturation-associated-inducible protein 1; Cold autoinflammatory syndrome 1 protein homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523919 275.00
Check your price
  • IDO Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA524598
  • Size 400 µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Indoleamine-pyrrole 2,3-dioxygenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524598 349.00
Check your price
  • VIP Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Bovine, Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # ABS0230202
  • Size 200 µL
Applications
ELISA (1:1000), Western Blot (Assay dependent)
Alternate names:
VIP-R-1, Pituitary adenylate cyclase-activating polypeptide type II receptor_x000D_ PACAP type II receptor, PACAP-R-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0230202 550.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Secondary Antibody, AP conjugate for Western Blot, IHC and IP
  • Host Goat
  • Target Species Rabbit
  • Conjugate AP
  • Catalog # 31346
  • Size 0.2 mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:50,000), Immunofluorescence (1:5,000-1:50,000), Immunohistochemistry (1:5,000-1:50,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:50,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31346 103.00
Check your price
  • Phospho-SHP-1 (Tyr564) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA537708
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
SH-PTP1; Protein-tyrosine phosphatase SHP-1; Protein-tyrosine phosphatase 1C; Hematopoietic cell protein-tyrosine phosphatase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537708 309.00
Check your price
  • DNAI1 Monoclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Chicken, Deer, Hamster, Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA122146
  • Size 100 µL
Applications
ELISA (Assay dependent), Immunocytochemistry (Assay dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:2000)
Alternate names:
Axonemal dynein intermediate chain 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122146 399.00
Check your price
  • Lysozyme Polyclonal Antibody for IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA126309
  • Size 6 mL
Applications
Immunohistochemistry (Frozen) (Neat), Immunohistochemistry (Paraffin) (Neat)
Alternate names:
1,4-beta-N-acetylmuramidase C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126309 309.00
Check your price
  • Phospho-c-Raf (Tyr341) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537713
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Raf-1; Proto-oncogene c-RAF
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537713 309.00
Check your price
  • beta-2 Adrenergic Receptor Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA527083
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Beta-2 adrenoreceptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527083 349.00
Check your price
  • Human Interleukin 4 (IL-4) for Western Blot, IP, ELISA and Ctrl
  • Host Yeast
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RIL4I
  • Size 20 ug
Applications
Control (Assay Dependent), ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Pitrakinra; Lymphocyte stimulatory factor 1; Binetrakin; B-cell stimulatory factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
RIL4I 309.00
Check your price
  • Cytokeratin Pan Monoclonal Antibody, APC conjugate for IF, ICC and Flow
  • Host Mouse
  • Target Species Mammal
  • Conjugate APC
  • Catalog # MA110325
  • Size 100 ug
Applications
Flow Cytometry (5 ug/ml), Immunocytochemistry (1:100-1:400), Immunofluorescence (1:100-1:400)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; Keratin-10; Cytokeratin-10; Type-II keratin Kb8; Keratin-8; Cytokeratin-8; Type-II keratin Kb5; Keratin-5; Cytokeratin-5; 58 kDa cytokeratin; Type-II keratin Kb6; Keratin-6A; Cytokeratin-6D; Cytokeratin-6A; Keratin-13; Cytokeratin-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110325 349.00
Check your price
  • Phospho-Estrogen Receptor alpha (Ser167) Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA537570
  • Size 100 µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537570 309.00
Check your price
  • SERCA2 ATPase Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P), Flow and IP
  • Host Mouse
  • Target Species Dog, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3919
  • Size 100 µL
Applications
Flow Cytometry (1µg per 10^6 cells), Immunocytochemistry (1:250), Immunofluorescence (1:100-1:200), Immunohistochemistry (1:100), Immunohistochemistry (Frozen) (1/100), Immunohistochemistry (Paraffin) (1 µg/ml), Immunoprecipitation (Assay dependent), Western Blot (1:1,000)
Alternate names:
Endoplasmic reticulum class 1/2 Ca(2+) ATPase; Calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletal muscle isoform; Calcium pump 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3919 375.00
Check your price
  • c-Fos Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSC00040W
  • Size 100 µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Cellular oncogene fos
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSC00040W 309.00
Check your price
  • CD44 Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517878
  • Size 100 ug
Applications
Flow Cytometry (0.5 ug/10^6 cells)
Alternate names:
Phagocytic glycoprotein I; Phagocytic glycoprotein 1; Lymphocyte antigen 24; Hyaluronate receptor; Hermes antigen; HUTCH-I; GP90 lymphocyte homing/adhesion receptor; Extracellular matrix receptor III
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517878 159.00
Check your price
  • Toxoplasma gondii SAG1 Monoclonal Antibody for Western Blot, IF and ELISA
  • Host Mouse
  • Target Species Protozoa
  • Conjugate Unconjugated
  • Catalog # MA518268
  • Size 100 µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50), Western Blot (1:10-1:50)
Alternate names:
T. gondii
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518268 286.00
Check your price
  • Goat anti-Mouse IgE Secondary Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA129273
  • Size 500 µg
Applications
ELISA (1-5 µg/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129273 302.00
Check your price
  • CD206 Monoclonal Antibody for Western Blot, IHC (F) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135936
  • Size 200 µg
Applications
Flow Cytometry (10ul x 10^6 cells or 100ul whole blood), Immunohistochemistry (Frozen) (Assay dependent.), Western Blot (Assay dependent.)
Alternate names:
Macrophage mannose receptor 1-like protein 1; Human mannose receptor; C-type lectin domain family 13 member D-like; C-type lectin domain family 13 member D
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135936 800.00
Check your price
  • Goat anti-Mouse IgG (H+L) Secondary Antibody, APC-Alexa Fluor® 750 conjugate for Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate APC-Alexa Fluor® 750
  • Catalog # A21006
  • Size 100 µg
Applications
Flow Cytometry (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21006 311.00
Check your price
  • Phospho-TTK (Thr360, Ser362, Ser363) Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 441330G
  • Size 10 blots
Applications
Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Phosphotyrosine picked threonine-protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
441330G 309.00
Check your price
  • Claudin 1 Polyclonal Antibody for Western Blot, IF, IHC (P) and ELISA
  • Host Rabbit
  • Target Species Chicken, Human, Rat
  • Conjugate Unconjugated
  • Catalog # 519000
  • Size 100 µg
Applications
ELISA (0.1-1.0 ug/ml), Immunofluorescence (5-10 ug/ml), Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (1-2 ug/ml)
Alternate names:
Senescence-associated epithelial membrane protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
519000 310.00
Check your price