Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  21  22  23  24  25  26  27  28  29  30  31  32  33  34  35  36  37  38  39  40  41  42  43  44  45  46  47  48  49  50  60  70  80  90  100  110  120  130  200  300  400  500  600  700  800  900  1000  1630 
1021 - 1050 of 48882 products displayed
  • PIR-A/PIR-B Monoclonal Antibody for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524140
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells)
Alternate names:
Paired immunoglobulin-like receptor B; Cell-surface glycoprotein p91
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524140 275.00
Check your price
  • Phospho-CaMKIV (Thr196, Thr200) Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538435
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
CaM kinase-GR; Calspermin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538435 309.00
Check your price
  • Donkey anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 568 conjugate for IF, ICC, IHC and Flow
  • Host Donkey
  • Target Species Goat
  • Conjugate Alexa Fluor® 568
  • Catalog # A11057
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11057 209.00
Check your price
  • PDGFRA Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # A18351
  • Size 50µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Platelet-derived growth factor alpha receptor; CD140 antigen-like family member A; Alpha-type platelet-derived growth factor receptor; Alpha platelet-derived growth factor receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18351 101.00
Check your price
  • Laminin gamma-1 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Bovine, Guinea pig, Human, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA126516
  • Size 500µL
Applications
Immunoprecipitation (1:400), Western Blot (1:200)
Alternate names:
S-laminin subunit gamma; Laminin-9 subunit gamma; Laminin-8 subunit gamma; Laminin-7 subunit gamma; Laminin-6 subunit gamma; Laminin-4 subunit gamma; Laminin-3 subunit gamma; Laminin-2 subunit gamma; Laminin-11 subunit gamma; Laminin-10 subunit gamma; Laminin-1 subunit gamma; Laminin B2 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126516 375.00
Check your price
  • IL-3 (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP75595
  • Size 1mg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75595 2,600.00
Check your price
  • Phospho-SNAIL (Ser246) Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA537739
  • Size 100µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Western Blot (1:500-1:1000)
Alternate names:
Protein snail homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537739 309.00
Check your price
  • SCD1 Monoclonal Antibody for Western Blot, IF, IHC and IP
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # A13996
  • Size 100µL
Applications
Immunofluorescence (1:100), Immunohistochemistry (1:200), Immunoprecipitation (Assay Dependent), Western Blot (1:1,000)
Alternate names:
Stearoyl-CoA desaturase 1; Fatty acid desaturase 1; Delta(9)-desaturase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
A13996 309.00
Check your price
  • GAPDH Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # TAB1001
  • Size 100ug
Applications
Western Blot (0.5 ug/ml)
Alternate names:
Peptidyl-cysteine S-nitrosylase GAPDH
Catalog # Size Price (AUD) Your Price (AUD) Qty
TAB1001 249.00
Check your price
  • beta-3 Tubulin Monoclonal Antibody for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Mouse
  • Target Species Bovine, Guinea pig, Hamster, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA1118X
  • Size 20µL
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1:50-1:200), Immunoprecipitation (5 µg), Western Blot (1:500-1:2000)
Alternate names:
Tubulin beta-III; Tubulin beta-4 chain; Neuron-specific class III beta-tubulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1118X 119.00
Check your price
  • TARDBP Polyclonal Antibody for Western Blot, IF, ICC, IHC (F) and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519712
  • Size 100µg
Applications
Immunocytochemistry (5 µg/ml), Immunofluorescence (5 µg/ml), Immunohistochemistry (Frozen) (Assay-Dependent), Immunoprecipitation (1:50), Western Blot (1 µg/ml)
Alternate names:
RP4-635E18.2, ALS10, TDP-43, TAR DNA-binding protein 43; TAR DNA-binding protein-43
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519712 349.00
Check your price
  • Defensin Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA182491
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (0.1-0.5 ug/ml), Immunohistochemistry (Paraffin) (0.2-1 ug/ml), Western Blot (Assay Dependent)
Alternate names:
HNP-4; Defensin, alpha 4; HNP-1; Defensin, alpha 1; HNP-3; Defensin, alpha 3; HD5(20-94); Defensin, alpha 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182491 800.00
Check your price
  • CD56 Monoclonal Antibody for IF, ICC and IHC (F)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA106800
  • Size 100µL
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (1:100-1:200)
Alternate names:
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106800 349.00
Check your price
  • CD24 Monoclonal Antibody, PE-Cyanine7 conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE-Cyanine7
  • Catalog # A14776
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
X62 heat stable antigen; R13-Ag; Nectadrin; M1/69-J11D heat stable antigen; Lymphocyte antigen 52
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14776 97.00
Check your price
  • Phospho-4E-BP1 (Ser65) Monoclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Chicken, Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA514948
  • Size 100µL
Applications
Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
Phosphorylated heat- and acid-stable protein regulated by insulin 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514948 309.00
Check your price
  • Jagged2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513136
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513136 349.00
Check your price
  • SLC5A5 Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121239
  • Size 500µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (1/50), Western Blot (1-2 µg/ml.)
Alternate names:
Solute carrier family 5 member 5; Sodium-iodide symporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121239 425.00
Check your price
  • 53BP1 Polyclonal Antibody, Biotin conjugate for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # PA522757
  • Size 100µl
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Western Blot (1:200-1:10,000)
Alternate names:
53BP1, p202, p53-binding protein 1, p53BP1, tumor protein 53-binding protein, 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522757 375.00
Check your price
  • GPR27 Polyclonal Antibody for IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532821
  • Size 50ug
Applications
ELISA (Assay-dependent), Immunohistochemistry (Paraffin) (10 µg/ml)
Alternate names:
Super conserved receptor expressed in brain 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532821 349.00
Check your price
  • VCAM-1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527217
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
INCAM-100
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527217 349.00
Check your price
  • F4/80 Monoclonal Antibody for IF, ICC, IHC, IHC (F), IHC (P), Flow, IP, RIA and IM
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA516624
  • Size 2mL
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunomicroscopy (Assay Dependent), Immunoprecipitation
Alternate names:
EMR1 hormone receptor; EGF-like module-containing mucin-like hormone receptor-like 1; EGF-like module receptor 1; Cell surface glycoprotein F4/80
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516624 475.00
Check your price
  • Cardiac Troponin I Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120122
  • Size 200µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Cardiac troponin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120122 309.00
Check your price
  • Aconitase 2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA511770
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Citrate hydro-lyase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511770 349.00
Check your price
  • VHL Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA146136
  • Size 100µL
Applications
Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
pVHL; Protein G7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146136 349.00
Check your price
  • Phospho-HBP1 (Ser402) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536809
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
High mobility group box transcription factor 1; HMG box transcription factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536809 349.00
Check your price
  • Phospho-p27 Kip1 (Thr187) Polyclonal Antibody for Western Blot, IF, IP and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 717700
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p27Kip1; Cyclin-dependent kinase inhibitor p27
Catalog # Size Price (AUD) Your Price (AUD) Qty
717700 305.00
Check your price
  • Trichomonas vaginalis Monoclonal Antibody for IF and ELISA
  • Host Mouse
  • Target Species Protozoa
  • Conjugate Unconjugated
  • Catalog # MA518273
  • Size 100µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50)
Alternate names:
T. vaginalis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518273 289.00
Check your price
  • Phospho-FAK2 (Tyr402) Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44618G
  • Size 10blots
Applications
Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (Assay Dependent)
Alternate names:
Related adhesion focal tyrosine kinase; Proline-rich tyrosine kinase 2; Focal adhesion kinase 2; Cell adhesion kinase beta; Calcium-regulated non-receptor proline-rich tyrosine kinase; Calcium-dependent tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
44618G 306.00
Check your price
  • PLZF Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529213
  • Size 100µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
Zinc finger protein PLZF; Zinc finger protein 145; Promyelocytic leukemia zinc finger protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529213 349.00
Check your price
  • EpoR Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523823
  • Size 100µg
Applications
ELISA (0.5-2 µg/ml (Detection), 2-8 µg/ml (Capture))
Alternate names:
EPO R, EPO-R, epor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523823 275.00
Check your price