Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  40  50  60  70  80  90  100  200  300  400  500  600  700  800  900  1000  1611 
91 - 120 of 48310 products displayed
  • IFN alpha Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 211002
  • Size 500 ug
Applications
ELISA (Assay Dependent), Neutralization (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Interferon alpha-D
Catalog # Size Price (AUD) Your Price (AUD) Qty
211002 499.00
Check your price
  • SF1 Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA536103
  • Size 100 µl
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Zinc finger protein 162; Zinc finger gene in MEN1 locus; Transcription factor ZFM1; Mammalian branch point-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536103 309.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 800 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate DyLight® 800
  • Catalog # SA510176
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510176 157.00
Check your price
  • CD31 Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514372
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
PECA1; GPIIA'; EndoCAM
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514372 349.00
Check your price
  • Rabbit anti-Bovine IgG (H+L) Secondary Antibody for ELISA
  • Host Rabbit
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # A24442
  • Size 2 mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24442 104.00
Check your price
  • BrdU Monoclonal Antibody, Alexa Fluor® 647 conjugate for Flow
  • Host Mouse
  • Target Species Chemical
  • Conjugate Alexa Fluor® 647
  • Catalog # B35140
  • Size 100 Tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
5-Bromo-2-deoxyuridine, Bromodeoxyuridine
Catalog # Size Price (AUD) Your Price (AUD) Qty
B35140 391.00
Check your price
  • CD14 Monoclonal Antibody, Alexa Fluor® 700 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Alexa Fluor® 700
  • Catalog # MHCD1429
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Myeloid cell-specific leucine-rich glycoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD1429 352.00
Check your price
  • GLUT1 Monoclonal Antibody for IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA511315
  • Size 500 µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Glucose transporter type 1, erythrocyte/brain; HepG2 glucose transporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511315 349.00
Check your price
  • PSMD4 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1966
  • Size 200 µL
Applications
Immunoprecipitation (Assay dependent), Western Blot (1:500)
Alternate names:
Multiubiquitin chain-binding protein; Antisecretory factor 1; 26S proteasome regulatory subunit S5A; 26S proteasome regulatory subunit RPN10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1966 349.00
Check your price
  • p21 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514456
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
p21; Melanoma differentiation-associated protein 6; CDK-interacting protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514456 349.00
Check your price
  • CD137 Monoclonal Antibody for IF and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513739
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent)
Alternate names:
T-cell antigen ILA; T-cell antigen 4-1BB homolog; CDw137; 4-1BB ligand receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513739 309.00
Check your price
  • PPAR alpha Monoclonal Antibody for Western Blot, IF, ICC, Flow, IP, ChIP and GS
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1822
  • Size 100 µg
Applications
ChIP assay (Assay Dependent), Flow Cytometry (1µg per 10^6 cells), Gel Shift (Assay dependent), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunoprecipitation (Assay dependent), Western Blot (2-5 µg/ml)
Alternate names:
Nuclear receptor subfamily 1 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1822 375.00
Check your price
  • LATS1/LATS2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539741
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Serine/threonine-protein kinase kpm; Large tumor suppressor homolog 2; Kinase phosphorylated during mitosis protein; WARTS protein kinase; Large tumor suppressor homolog 1; Warts-like kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539741 309.00
Check your price
  • Actin Muscle Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA514084
  • Size 500 µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:100-1:500)
Alternate names:
Smooth muscle gamma-actin; Gamma-2-actin; Alpha-actin-3; Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-cardiac actin; Alpha-actin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514084 309.00
Check your price
  • Goat anti-Mouse IgG1 Secondary Antibody, Alexa Fluor® 594 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate
  • Catalog # A21125
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21125 149.00
Check your price
  • Goat anti-Human IgG Fc Cross-Adsorbed Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # A18827
  • Size 2 mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18827 147.00
Check your price
  • DR5 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA119759
  • Size 0.1 mg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
TNF-related apoptosis-inducing ligand receptor 2; Death receptor 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119759 309.00
Check your price
  • Avidin Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Chicken
  • Conjugate Unconjugated
  • Catalog # MA121418
  • Size 500 µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
AVD, avID
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121418 309.00
Check your price
  • SLC25A20 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Sheep, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA542411
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Solute carrier family 25 member 20; Carnitine/acylcarnitine translocase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542411 309.00
Check your price
  • Human IgG Isotype Control for IF, ICC, IHC, BLOCK and Ctrl
  • Host Human
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # 31154
  • Size 10 mg
Applications
Blocking Assay (Assay Dependent), Control (Assay-Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
Negative Control, Isotype Control, Immunoglobulin Control, Control
Catalog # Size Price (AUD) Your Price (AUD) Qty
31154 149.00
Check your price
  • Myosin Skeletal Muscle Monoclonal Antibody for Western Blot, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Mammal
  • Conjugate Unconjugated
  • Catalog # MA135718
  • Size 1 mL
Applications
Immunohistochemistry (Frozen) (1:5-1:10), Immunohistochemistry (Paraffin) (1:5-1:10), Western Blot (Assay Dependent)
Alternate names:
Myosin heavy chain, skeletal muscle, adult 1; Myosin heavy chain 2x; Myosin heavy chain 1; Myosin heavy chain IIx/d
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135718 399.00
Check your price
  • Parkin Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513398
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Parkinson juvenile disease protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513398 349.00
Check your price
  • Donkey anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Donkey
  • Target Species Rabbit
  • Conjugate Unconjugated
  • Catalog # 31238
  • Size 1 mg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31238 113.00
Check your price
  • Human Interleukin 4 (IL-4) for Western Blot, IP, ELISA and Ctrl
  • Host Yeast
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RIL4MI
  • Size 1 mg
Applications
Control (Assay Dependent), ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Pitrakinra; Lymphocyte stimulatory factor 1; Binetrakin; B-cell stimulatory factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
RIL4MI 3,625.00
Check your price
  • CD147 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # A15772
  • Size 100 tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Tumor cell-derived collagenase stimulatory factor; OK blood group antigen; Leukocyte activation antigen M6; Extracellular matrix metalloproteinase inducer; Collagenase stimulatory factor; 5F7
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15772 171.00
Check your price
  • Cytokeratin Pan Monoclonal Antibody for Western Blot, IF, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135754
  • Size 1 mL
Applications
Immunofluorescence (Assay dependent.), Immunohistochemistry (Paraffin, Frozen) ((1:10, 1:50)), Western Blot (Assay dependent.)
Alternate names:
Type-II keratin Kb1; Keratin-1; Hair alpha protein; Cytokeratin-1; 67 kDa cytokeratin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135754 309.00
Check your price
  • Neisseria gonorrhoeae Polyclonal Antibody, Biotin conjugate for IF, ICC and ELISA
  • Host Rabbit
  • Target Species Bacteria
  • Conjugate Biotin
  • Catalog # PA127386
  • Size 100 µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent)
Alternate names:
N. gonorrhoeae, Gonococci
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127386 309.00
Check your price
  • RBP4 Monoclonal Antibody for Western Blot and Flow
  • Host Rat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA524158
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.05-2 µg/ml)
Alternate names:
Plasma retinol-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524158 275.00
Check your price
  • HMGA2 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), IP and ChIP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521320
  • Size 100 µL
Applications
ChIP assay (Assay-dependent), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:5000-1:20,000)
Alternate names:
High mobility group AT-hook protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521320 349.00
Check your price
  • EpCAM Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA110197
  • Size 100 Tests
Applications
Flow Cytometry (20 ul/10^6 cells)
Alternate names:
Tumor-associated calcium signal transducer 1; Major gastrointestinal tumor-associated protein GA733-2; KSA; KS 1/4 antigen; Epithelial glycoprotein 314; Epithelial glycoprotein; Epithelial cell surface antigen; Cell surface glycoprotein Trop-1; Adenocarcinoma-associated antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110197 249.00
Check your price