Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  33  34  35  36  37  38  39  40  41  42  43  44  45  46  47  48  49  50  51  52  53  54  55  56  57  58  59  60  61  62  70  80  90  100  110  120  130  140  200  300  400  500  600  700  800  900  1000  1630 
1411 - 1440 of 48881 products displayed
  • AKR1B10 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P), IP and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523017
  • Size 100µL
Applications
ELISA (1:100-1:2000), Immunocytochemistry (1:5-1:50), Immunofluorescence (1:5-1:50), Immunohistochemistry (Frozen) (1:5-1:50), Immunohistochemistry (Paraffin) (1:5-1:50), Immunoprecipitation (5 µg), Western Blot (1:500-1:1000)
Alternate names:
Small intestine reductase; Aldose reductase-related protein; Aldose reductase-like; ARL-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523017 349.00
Check your price
  • GST Tag Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # CAB4169
  • Size 100ug
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Glutathione-S-Transferase, GST protein, GST Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
CAB4169 149.00
Check your price
  • Entamoeba histolytica Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Protozoa
  • Conjugate Unconjugated
  • Catalog # MA110659
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
E. histolytica
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110659 349.00
Check your price
  • PD-L1 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # A27087
  • Size 100µg
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
B7 homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
A27087 153.00
Check your price
  • CD44 Monoclonal Antibody, PE-Cyanine5 conjugate for Flow
  • Host Rat
  • Target Species Human, Mouse
  • Conjugate PE-Cyanine5
  • Catalog # A16218
  • Size 50µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Phagocytic glycoprotein I; Phagocytic glycoprotein 1; Lymphocyte antigen 24; Hyaluronate receptor; Hermes antigen; HUTCH-I; GP90 lymphocyte homing/adhesion receptor; Extracellular matrix receptor III; Heparan sulfate proteoglycan; Epican; CDw44
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16218 149.00
Check your price
  • RPTN Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539412
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539412 309.00
Check your price
  • BNIP3L Monoclonal Antibody for Western Blot, IF, ICC, Flow and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # 393300
  • Size 100µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
NIP3-like protein X; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3A; Adenovirus E1B19K-binding protein B5
Catalog # Size Price (AUD) Your Price (AUD) Qty
393300 304.00
Check your price
  • CLEC2I Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547481
  • Size 100µg
Applications
Western Blot (0.1 µg/ml)
Alternate names:
Osteoclast inhibitory lectin-related protein 2; Lymphoid-derived C-type lectin-1; C-type lectin-related protein G; C-type lectin-related protein DCL1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547481 349.00
Check your price
  • Aspergillus niger Glucose Oxidase Polyclonal Antibody for ELISA
  • Host Rabbit
  • Target Species Fungi
  • Conjugate Unconjugated
  • Catalog # PA185473
  • Size 2mL
Applications
ELISA (Assay Dependent)
Alternate names:
A. niger Glucose Oxidase, Aspergillus niger GOx, A. niger GOx
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185473 319.00
Check your price
  • Human Interleukin 6 (IL-6) for Western Blot, IP, ELISA and Ctrl
  • Host Yeast
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RIL6I
  • Size 20ug
Applications
Control (Assay Dependent), ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Interferon beta-2; Hybridoma growth factor; CTL differentiation factor; B-cell stimulatory factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
RIL6I 308.00
Check your price
  • Caspase 8 (Cleaved Asp391) Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515054
  • Size 100µL
Applications
Flow Cytometry (1:100), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:100), Western Blot (1:1000)
Alternate names:
MORT1-associated ced-3 homolog; ICE-like apoptotic protease 5; FADD-like ICE; FADD-homologous ICE/ced-3-like protease; CAP4; Apoptotic protease Mch-5; Apoptotic cysteine protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515054 309.00
Check your price
  • Phospho-ZAP70 (Tyr315, Tyr319) Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 700177
  • Size 100µg
Applications
Flow Cytometry (1-3 ug/test), Immunocytochemistry (1-3 ug/ml), Immunofluorescence (1-3 ug/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
Syk-related tyrosine kinase; 70 kDa zeta-chain associated protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
700177 303.00
Check your price
  • CD206 Monoclonal Antibody for IHC (F)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516546
  • Size 200ug
Applications
Immunohistochemistry (Frozen) (1:50-1:500)
Alternate names:
Macrophage mannose receptor 1-like protein 1; Human mannose receptor; C-type lectin domain family 13 member D-like; C-type lectin domain family 13 member D
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516546 399.00
Check your price
  • Desmoplakin Monoclonal Antibody for Western Blot, IF and IHC (F)
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA183118
  • Size 50µg
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
250/210 kDa paraneoplastic pemphigus antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183118 475.00
Check your price
  • CD63 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 10628D
  • Size 200µL
Applications
Western Blot (Assay Dependent)
Alternate names:
Tetraspanin-30; Ocular melanoma-associated antigen; OMA81H; Melanoma-associated antigen ME491; Lysosomal-associated membrane protein 3; Granulophysin
Catalog # Size Price (AUD) Your Price (AUD) Qty
10628D 234.00
Check your price
  • LC3B Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ChIP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 700712
  • Size 100µg
Applications
ChIP assay (1 ul), Flow Cytometry (3-5 ug x 10^6 cells), Immunocytochemistry (1-3 ug/ml), Immunofluorescence (1-3 ug/ml), Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (1-5 ug/ml)
Alternate names:
Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
700712 303.00
Check your price
  • Erythrocytes Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate Biotin
  • Catalog # MA517578
  • Size 100ug
Applications
Flow Cytometry (1.0 ug/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517578 159.00
Check your price
  • Peptidoglycan Monoclonal Antibody for IHC (F), IHC (P) and ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA516509
  • Size 100µL
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516509 425.00
Check your price
  • Smooth Muscle Actin Monoclonal Antibody for Western Blot, ICC, IHC, ELISA and IM
  • Host Mouse
  • Target Species Chicken, Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA112772
  • Size 50µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (1:1000-1:10000), Immunomicroscopy (Assay Dependent), Western Blot (1:10000-1:50000)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112772 425.00
Check your price
  • ICAM-1 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # A18679
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Major group rhinovirus receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18679 125.00
Check your price
  • c-Mpl Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517127
  • Size 100ug
Applications
Flow Cytometry (1:200-1:400), Western Blot (1:500-1:2000)
Alternate names:
Proto-oncogene c-Mpl; Myeloproliferative leukemia protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517127 309.00
Check your price
  • Estrogen Receptor alpha Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Hamster, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA516440
  • Size 500µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:250-500), Immunoprecipitation (10 µg/ml), Western Blot (2-5µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516440 349.00
Check your price
  • SLC7A7 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA542432
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
y(+)L-type amino acid transporter 1; Solute carrier family 7 member 7; Monocyte amino acid permease 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542432 309.00
Check your price
  • VEGF Polyclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # P931
  • Size 500ug
Applications
ELISA (Assay Dependent), Immunocytochemistry (1:20-1:100), Immunofluorescence (1:20-1:100), Western Blot (1:500-1:1000)
Alternate names:
Vascular permeability factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
P931 375.00
Check your price
  • GnRHR Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518623
  • Size 100ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
GNRHR; LRHR; gonadotropin-releasing hormone receptor; GNRHR1; GRHR; LHRHR; gonadotropin-releasing hormone (type 1) receptor 1; leutinizing hormone releasing horomone receptor; leutinizing-releasing hormone receptor; luliberin receptor; type I GnRH receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518623 349.00
Check your price
  • ACTH Monoclonal Antibody for Western Blot, IF, ICC, Flow and ELISA
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 701293
  • Size 100µg
Applications
ELISA (Assay-dependent), Western Blot (1-3 ug/ml)
Alternate names:
Corticotropin-lipotropin
Catalog # Size Price (AUD) Your Price (AUD) Qty
701293 309.00
Check your price
  • Rabbit anti-Sheep IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Sheep
  • Conjugate HRP
  • Catalog # 31480
  • Size 1.5mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:100,000), Immunofluorescence (1:5,000-1:100,000), Immunohistochemistry (1:5,000-1:100,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:200,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31480 149.00
Check your price
  • ADAM10 Monoclonal Antibody for Western Blot and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523867
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Mammalian disintegrin-metalloprotease; Kuzbanian protein homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523867 275.00
Check your price
  • STK33 Polyclonal Antibody for Western Blot, IF, ICC, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523081
  • Size 100µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Paraffin) (1:100), Western Blot (1:20,000)
Alternate names:
serine/threonine-protein kinase 33
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523081 349.00
Check your price
  • Legionella pneumophila Polyclonal Antibody, FITC conjugate for IF
  • Host Rabbit
  • Target Species Bacteria
  • Conjugate FITC
  • Catalog # PA173140
  • Size 1mL
Applications
Immunofluorescence (1:10-1:50)
Alternate names:
L. pneumophila, Legionellosis, Legionnaires disease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173140 375.00
Check your price