Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  41  42  43  44  45  46  47  48  49  50  51  52  53  54  55  56  57  58  59  60  61  62  63  64  65  66  67  68  69  70  80  90  100  110  120  130  140  150  200  300  400  500  600  700  800  900  1000  1611 
1651 - 1680 of 48310 products displayed
  • IGF2R Antibody, Alexa Fluor® 647 (Monoclonal, MEM-238)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Alexa Fluor® 647
  • Catalog # MA518132
  • Size 100 tests
Applications
Flow Cytometry (4ulx100ul whole blood or 10^6 cells)
Alternate names:
M6P/IGF2 receptor; Insulin-like growth factor II receptor; Insulin-like growth factor 2 receptor; 300 kDa mannose 6-phosphate receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518132 249.00
Check your price
  • SOD1 Antibody (Monoclonal, 6F5)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515520
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Western Blot (1/500-1/2000)
Alternate names:
Superoxide dismutase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515520 349.00
Check your price
  • CD40 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532325
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
CDw40; CD40L receptor; Bp50; B-cell surface antigen CD40
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532325 275.00
Check your price
  • Phospho-PKR (Thr451) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538145
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
p68 kinase; Tyrosine-protein kinase EIF2AK2; Serine/threonine-protein kinase TIK; Protein kinase RNA-activated; P1/eIF-2A protein kinase; Interferon-inducible RNA-dependent protein kinase; Eukaryotic translation initiation factor 2-alpha kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538145 309.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed, Alexa Fluor® 568
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 568
  • Catalog # A11004
  • Size 500 µL
Applications
Flow Cytometry (1-10 µg/mL), Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11004 169.00
Check your price
  • Collagen III Antibody
  • Host Rabbit
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # PA128870
  • Size 100 µg
Applications
ELISA (1:5000-1:10,000), Immunohistochemistry (Frozen) (1:50-1:200), Immunohistochemistry (Paraffin) (Assay-Dependent), Immunoprecipitation (Assay dependent), Western Blot (1:5000-1:10,000)
Alternate names:
COL3, Collagen type III alpha 1, EDS4A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128870 309.00
Check your price
  • OPRK1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124534
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124534 425.00
Check your price
  • Phospho-RPA2 (Ser33) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539809
  • Size 100 µg
Applications
peptide-ELISA (1:10,000), Western Blot (1:500-1:1000)
Alternate names:
Replication protein A 34 kDa subunit; Replication factor A protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539809 309.00
Check your price
  • Neutrophil elastase Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529659
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
PMN elastase; Medullasin; Human leukocyte elastase; Elastase-2; Bone marrow serine protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529659 349.00
Check your price
  • Epo Antibody (Monoclonal, 148436)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523874
  • Size 100 µg
Applications
ELISA (0.5-2 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.15-0.75 µg/ml)
Alternate names:
EP, MGC138142, MVCD2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523874 275.00
Check your price
  • Goat anti-Rabbit IgG, Alexa Fluor® 488-5 nm colloidal gold
  • Host Goat
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 488-5 nm colloidal gold
  • Catalog # A31565
  • Size 500 µL
Applications
Immunomicroscopy (0.2-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A31565 209.00
Check your price
  • EpCAM Antibody (Monoclonal, 323/A3)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA512436
  • Size 500 µL
Applications
Flow Cytometry (0.5-1 ug/test), Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (1-2 µg/ml), Radioimmune Assays (Assay Dependent)
Alternate names:
Tumor-associated calcium signal transducer 1; Protein 289A; Epithelial glycoprotein 314; Major gastrointestinal tumor-associated protein GA733-2; KSA; KS 1/4 antigen; Epithelial glycoprotein; Epithelial cell surface antigen; Cell surface glycoprotein Trop-1; Adenocarcinoma-associated antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512436 309.00
Check your price
  • SLC26A6 Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA537971
  • Size 100 µg
Applications
Western Blot (0.1-0.3 µg/ml)
Alternate names:
Putative anion transporter-1; Pendrin-like protein 1; Pendrin-L1; Chloride-formate exchanger; Anion exchange transporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537971 309.00
Check your price
  • SUGT1 Antibody (Monoclonal, 1A10)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA521189
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Immunoprecipitation (Assay-Dependent), Western Blot (1-5 µg/ml), Western Blot (Assay-Dependent)
Alternate names:
Suppressor of G2 allele of SKP1 homolog; Sgt1; Protein 40-6-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA521189 309.00
Check your price
  • Hepatitis B Virus Surface Ad/Ay Antibody, FITC
  • Host Rabbit
  • Target Species Virus
  • Conjugate FITC
  • Catalog # PA173086
  • Size 1 mL
Applications
Immunofluorescence (1:10)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg, HBVgp2, HepB ad, HepB ay
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173086 309.00
Check your price
  • Salmonella LPS Antibody (Monoclonal, A99H)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA518258
  • Size 100 µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50)
Alternate names:
Salmonellae
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518258 289.00
Check your price
  • PAP Antibody (Monoclonal, P1D8)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIP0101
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIP0101 600.00
Check your price
  • Goat anti-Rat IgG Fc, HRP
  • Host Goat
  • Target Species Rat
  • Conjugate HRP
  • Catalog # 31475
  • Size 2 mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:100,000), Immunofluorescence (1:5,000-1:100,000), Immunohistochemistry (1:5,000-1:100,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:200,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31475 179.00
Check your price
  • HEXA Antibody (Monoclonal, 714729)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524335
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (8-25 µg/ml), Western Blot (2 µg/ml)
Alternate names:
N-acetyl-beta-glucosaminidase subunit alpha; Beta-N-acetylhexosaminidase subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524335 275.00
Check your price
  • PPL Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530931
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
195 kDa cornified envelope precursor protein; 190 kDa paraneoplastic pemphigus antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530931 349.00
Check your price
  • Kanamycin Antibody
  • Host Sheep
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # PA175225
  • Size 100 µL
Applications
ELISA (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175225 600.00
Check your price
  • Phospho-MCL1 (Ser159, Thr163) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517125
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
mcl1/EAT; Bcl-2-related protein EAT/mcl1; Bcl-2-like protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517125 309.00
Check your price
  • PST1 Antibody
  • Host Rabbit
  • Target Species Yeast
  • Conjugate Unconjugated
  • Catalog # PA534414
  • Size 100 ug
Applications
ELISA (1 ug/mL), Western Blot (1 ug/mL)
Alternate names:
Protoplast secreted protein 1; Haze protective factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534414 349.00
Check your price
  • Luteinizing Hormone Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532478
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:500)
Alternate names:
Thyrotropin alpha chain; Thyroid-stimulating hormone alpha chain; Lutropin alpha chain; Luteinizing hormone alpha chain; Follitropin alpha chain; Follicle-stimulating hormone alpha chain; Chorionic gonadotrophin subunit alpha; Choriogonadotropin alpha chain; Anterior pituitary glycoprotein hormones common subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532478 349.00
Check your price
  • Mycoplasma pneumoniae Antibody (Monoclonal, S64D)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA518233
  • Size 100 µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50), Western Blot (1:10-1:50)
Alternate names:
bacterial pneumonia, M. pneumoniae
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518233 287.00
Check your price
  • Cytokeratin 8 Antibody (Monoclonal, SP102)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516420
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516420 499.00
Check your price
  • Trypsin Antibody (Monoclonal, TRYP11-236.7)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110617
  • Size 500 µg
Applications
ELISA (Assay dependent)
Alternate names:
Trypsin I; Serine protease 1; Cationic trypsinogen; Beta-trypsin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110617 349.00
Check your price
  • SSEA4 Antibody, Alexa Fluor® 647 (Monoclonal, MC813-70)
  • Host Mouse
  • Target Species Human
  • Conjugate Alexa Fluor® 647
  • Catalog # SSEA421
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
stage-specific embryonic antigen 4, SSEA4
Catalog # Size Price (AUD) Your Price (AUD) Qty
SSEA421 322.00
Check your price
  • CD4 Antibody, FITC (Monoclonal, YTS191.1)
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517442
  • Size 100 ug
Applications
Flow Cytometry (0.2 ug/10^6 cells)
Alternate names:
T-cell surface antigen T4/Leu-3; T-cell differentiation antigen L3T4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517442 119.00
Check your price
  • Phospho-SHIP2 (Tyr497) Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523331
  • Size 20 ug
Applications
Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (0.01-1 µg/ml)
Alternate names:
SH2 domain-containing inositol 5'-phosphatase 2; Protein 51C; Inositol polyphosphate phosphatase-like protein 1; AblSH3-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523331 349.00
Check your price