Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  45  46  47  48  49  50  51  52  53  54  55  56  57  58  59  60  61  62  63  64  65  66  67  68  69  70  71  72  73  74  80  90  100  110  120  130  140  150  200  300  400  500  600  700  800  900  1000  1630 
1771 - 1800 of 48877 products displayed
  • CD200R Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA135932
  • Size 100µg
Applications
Flow Cytometry (10ul x 10^6 cells or 100ul whole blood)
Alternate names:
Cell surface glycoprotein OX2 receptor 1; CD200 cell surface glycoprotein receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135932 600.00
Check your price
  • SMAD2 Monoclonal Antibody for Western Blot, IF, ICC, Flow, ChIP and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 700048
  • Size 100µg
Applications
ChIP assay (3µg), ELISA (1-5 ug/ml), Flow Cytometry (1-3µg x 10^6 cells), Immunocytochemistry (1µg/ml), Immunofluorescence (1µg/ml), Western Blot (0.5-1µg/ml)
Alternate names:
SMAD family member 2; Mad-related protein 2; JV18-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
700048 309.00
Check your price
  • PLVAP Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531402
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Plasmalemma vesicle protein 1; Fenestrated endothelial-linked structure protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531402 349.00
Check your price
  • Cytokeratin Pan Monoclonal Antibody for Western Blot, IF, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135754
  • Size 1mL
Applications
Immunofluorescence (Assay dependent.), Immunohistochemistry (Paraffin, Frozen) ((1:10, 1:50)), Western Blot (Assay dependent.)
Alternate names:
Type-II keratin Kb1; Keratin-1; Hair alpha protein; Cytokeratin-1; 67 kDa cytokeratin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135754 309.00
Check your price
  • CD8 alpha Monoclonal Antibody, Alexa Fluor® 700 conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 700
  • Catalog # MCD0824
  • Size 1mL
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
T-cell surface glycoprotein Lyt-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MCD0824 305.00
Check your price
  • GYS1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529739
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
GSY, GYS, glycogen [starch] synthase, muscle
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529739 349.00
Check your price
  • GFAP Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA135377
  • Size 1mL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Immunohistochemistry (Paraffin, Frozen) (1:10-1:50), Western Blot (Assay dependent)
Alternate names:
GFAP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135377 309.00
Check your price
  • Phospho-PPAR alpha (Ser12) Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1820
  • Size 100µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:100-1:1000)
Alternate names:
Nuclear receptor subfamily 1 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1820 375.00
Check your price
  • Phospho-TrkB (Tyr705) Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538077
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
TrkB tyrosine kinase; Neurotrophic tyrosine kinase receptor type 2; Tropomyosin-related kinase B; GP145-TrkB; GP145-TrkB/GP95-TrkB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538077 309.00
Check your price
  • MMP2 Monoclonal Antibody for IF, ICC, IHC, IHC (F), IHC (P), IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA116641
  • Size 100µL
Applications
ELISA (0.03 µg/ml), Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Frozen) (2 µg/ml), Immunohistochemistry (Paraffin) (2 µg/ml), Immunoprecipitation (1-2 µg/100-500 µg total protein), Western Blot (1-2 µg/ml)
Alternate names:
TBE-1; Matrix metalloproteinase-2; Gelatinase A; 72 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116641 375.00
Check your price
  • Nalidixic Acid Polyclonal Antibody for ELISA
  • Host Sheep
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # PA175301
  • Size 100µL
Applications
ELISA (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175301 650.00
Check your price
  • Phospho-DNA-PK (Thr2609) Polyclonal Antibody for DB
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512913
  • Size 400µL
Applications
Dot blot (1:500)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512913 349.00
Check your price
  • SIGLEC11 Polyclonal Antibody for Western Blot, Flow and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA548029
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
UNQ9222/PRO28718, sialic acid-binding Ig-like lectin 11, siglec-11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA548029 309.00
Check your price
  • CD103 Monoclonal Antibody for IHC (F), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121434
  • Size 100µg
Applications
Flow Cytometry (2 µg for 5x10^5 cells), Immunohistochemistry (Frozen) (1:50-1:100), Immunoprecipitation (Assay dependent)
Alternate names:
Mucosal lymphocyte 1 antigen; Integrin alpha-IEL; HML-1 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121434 349.00
Check your price
  • Goat anti-Rabbit IgG Secondary Antibody, Alexa Fluor® 488-10 nm colloidal gold conjugate for IM
  • Host Goat
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 488-10 nm colloidal gold
  • Catalog # A31566
  • Size 500µL
Applications
Immunomicroscopy (0.2-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A31566 209.00
Check your price
  • Acetyl-Histone H3 (Lys56) Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A16634
  • Size 100µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16634 Not Orderable Online
Check your price
  • Amodiaquine Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # HYB3200402
  • Size 200µl
Applications
ELISA (1:4,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB3200402 425.00
Check your price
  • CXCL8 Monoclonal Antibody for Western Blot, Flow and ELISA
  • Host Mouse
  • Target Species Bovine, Dog, Ferret, Mustelid, Mink, Sheep, Pig, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA182421
  • Size 250µg
Applications
ELISA (5 ug/ml), Flow Cytometry (1/10), Western Blot (Assay Dependent)
Alternate names:
Chemokine (C-X-C motif) ligand 8; C-X-C motif chemokine 8; Permeability factor 1; Neutrophil attractant/activation protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182421 650.00
Check your price
  • HTR1F Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533299
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (5-8 µg/ml)
Alternate names:
Serotonin receptor 1F
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533299 349.00
Check your price
  • beta Tubulin Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 480011
  • Size 100µL
Applications
Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (1:10000)
Alternate names:
Tubulin beta-5 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
480011 308.00
Check your price
  • Human Interleukin 1 beta (IL-1b) for Western Blot, IP, ELISA and Ctrl
  • Host Yeast
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RIL1BI
  • Size 20ug
Applications
Control (Assay Dependent), ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
RIL1BI 310.00
Check your price
  • IL17RC Polyclonal Antibody for Western Blot, Flow and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547286
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Neutralization (0.6-3.6 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
ZcytoR14; Interleukin-17 receptor-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547286 349.00
Check your price
  • SSR2 Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA520028
  • Size 100µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
Signal sequence receptor subunit beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA520028 309.00
Check your price
  • Nanog Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA1017
  • Size 100µg
Applications
Flow Cytometry (Assay dependent), Immunocytochemistry (1:50 -1:100), Immunofluorescence (1:50 -1:100), Western Blot (1:500-1:2000)
Alternate names:
Homeobox transcription factor Nanog
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1017 249.00
Check your price
  • ZO-1 Monoclonal Antibody, Alexa Fluor® 488 conjugate for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Dog, Human
  • Conjugate Alexa Fluor® 488
  • Catalog # 339188
  • Size 100µg
Applications
ELISA (0.1-1.0 ug/ml), Immunocytochemistry (Assay Dependent), Immunofluorescence (5-10 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Zonula occludens protein 1; Zona occludens protein 1; Tight junction protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
339188 359.00
Check your price
  • Nkx2.5 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523387
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Homeobox protein NK-2 homolog E; Homeobox protein CSX; Cardiac-specific homeobox
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523387 349.00
Check your price
  • Goat anti-Hamster IgG Cross-Adsorbed Secondary Antibody, HRP conjugate for ELISA
  • Host Goat
  • Target Species Hamster
  • Conjugate HRP
  • Catalog # PA533286
  • Size 800ug
Applications
ELISA (1:1000-1:5000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533286 375.00
Check your price
  • CD5 Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517045
  • Size 100ug
Applications
Flow Cytometry (1:200-1:400), Western Blot (1:250)
Alternate names:
Lymphocyte antigen T1/Leu-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517045 309.00
Check your price
  • ABCC9 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Goat, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA542398
  • Size 100µl
Applications
Western Blot (1:1000)
Alternate names:
Sulfonylurea receptor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542398 309.00
Check your price
  • beta Actin Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), IP, ELISA and Neu
  • Host Rabbit
  • Target Species Avian, Fish, Hamster, Human, Mouse, Non-human primate, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA146296
  • Size 50µL
Applications
Blocking Assay (Assay-Dependent), ELISA (Assay-Dependent), Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:10-1:500), Neutralization (Assay-Dependent), Western Blot (1:100-1:2000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146296 375.00
Check your price