Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  46  47  48  49  50  51  52  53  54  55  56  57  58  59  60  61  62  63  64  65  66  67  68  69  70  71  72  73  74  75  80  90  100  110  120  130  140  150  160  200  300  400  500  600  700  800  900  1000  1630 
1801 - 1830 of 48882 products displayed
  • alpha Tubulin Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 322500
  • Size 100µg
Applications
Immunocytochemistry (5-10 µg/ml), Immunofluorescence (5-10 ug/ml), Western Blot (1-3 ug/ml)
Alternate names:
Tubulin alpha-4 chain; Alpha-tubulin 4; Alpha-tubulin isotype M-alpha-4; Tubulin alpha-1 chain; Tubulin H2-alpha; Testis-specific alpha-tubulin; Alpha-tubulin 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
322500 307.00
Check your price
  • HLA-DR/DP/DQ Monoclonal Antibody for IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA125910
  • Size 6mL
Applications
Immunohistochemistry (Frozen) (Neat), Immunohistochemistry (Paraffin) (Neat)
Alternate names:
MHC class II antigen DPB1; HLA class II histocompatibility antigen, DP(W4) beta chain; MHC class II antigen DRB1*15; DW2.2/DR2.2; MHC class II antigen DRB1*16; MHC class II antigen DQB1; MHC class II antigen DRB1*3; Clone P2-beta-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125910 349.00
Check your price
  • Caspase 3 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 700182
  • Size 100µg
Applications
Flow Cytometry (0.5-1 ug/test), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (0.1-0.2 ug/ml)
Alternate names:
SREBP cleavage activity 1; Protein Yama; Cysteine protease CPP32; Apopain; LICE
Catalog # Size Price (AUD) Your Price (AUD) Qty
700182 309.00
Check your price
  • Tet Repressor Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # PA14501
  • Size 100µg
Applications
ELISA (Assay dependent), Western Blot (1:1000)
Alternate names:
TetR, pSC101 tetR, Tet Repressor Protein, Tetracycline Repressor Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14501 375.00
Check your price
  • CD36 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA119772
  • Size 100tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Thrombospondin receptor; Platelet glycoprotein IV; Platelet collagen receptor; PAS-4; PAS IV; Leukocyte differentiation antigen CD36; Glycoprotein IIIb; Fatty acid translocase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119772 309.00
Check your price
  • Armenian Hamster IgG Isotype Control for Flow and Ctrl
  • Host Hamster
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # PA533219
  • Size 250ug
Applications
Control (Assay-Dependent), Flow Cytometry (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533219 225.00
Check your price
  • Villin Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529078
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:5000-1:20,000)
Alternate names:
D2S1471, VIL, villin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529078 349.00
Check your price
  • CD24 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MHCD2404
  • Size 0.5ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Small cell lung carcinoma cluster 4 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD2404 265.00
Check your price
  • Hepatitis B Virus Surface Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MIH9705
  • Size 1mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9705 600.00
Check your price
  • TMEM27 Polyclonal Antibody for Western Blot and IHC (F)
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547676
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (1-10 µg/ml)
Alternate names:
Transmembrane protein 27
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547676 349.00
Check your price
  • SSRP1 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA112699
  • Size 100µg
Applications
Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
T160; Structure-specific recognition protein 1; Recombination signal sequence recognition protein 1; Facilitates chromatin transcription complex subunit SSRP1; Facilitates chromatin transcription complex 80 kDa subunit; Chromatin-specific transcription elongation factor 80 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112699 309.00
Check your price
  • Ataxin 3 Polyclonal Antibody for IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512088
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100)
Alternate names:
Spinocerebellar ataxia type 3 protein; Machado-Joseph disease protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512088 349.00
Check your price
  • Donkey anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, FITC conjugate for IF and Flow
  • Host Donkey
  • Target Species Goat
  • Conjugate FITC
  • Catalog # A16006
  • Size 1mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16006 125.00
Check your price
  • FXR1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529853
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
FXR1P, fragile X mental retardation syndrome-related protein 1, hFXR1p
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529853 349.00
Check your price
  • Donkey anti-Goat IgG Secondary Antibody, Texas Red® conjugate for IF, ICC, Flow and ELISA
  • Host Donkey
  • Target Species Goat
  • Conjugate Texas Red®
  • Catalog # PA128662
  • Size 1mg
Applications
Flow Cytometry (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128662 196.00
Check your price
  • SOCS3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # A14000
  • Size 100µL
Applications
Western Blot (1:1,000)
Alternate names:
Cytokine-inducible SH2 protein 3; STAT-induced STAT inhibitor 3; Protein EF-10
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14000 309.00
Check your price
  • ZO-1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519090
  • Size 100µg
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Zonula occludens protein 1; Zona occludens protein 1; Tight junction protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519090 349.00
Check your price
  • CD3 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate Biotin
  • Catalog # MA516460
  • Size 100ug
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516460 225.00
Check your price
  • Phospho-AMPK alpha-1 (Thr172) Monoclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701068
  • Size 100µg
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Western Blot (2-5 ug/ml)
Alternate names:
Tau-protein kinase PRKAA1; Hydroxymethylglutaryl-CoA reductase kinase; Acetyl-CoA carboxylase kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
701068 309.00
Check your price
  • CUX1 Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530003
  • Size 100µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunoprecipitation (1:100-1:500), Western Blot (1:500-1:3000)
Alternate names:
Homeobox protein cux-1; CCAAT displacement protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530003 349.00
Check your price
  • TNF alpha Polyclonal Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Rabbit
  • Target Species Bovine, Sheep
  • Conjugate Biotin
  • Catalog # PBOTNFABI
  • Size 250ug
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Tumor necrosis factor ligand superfamily member 2; TNF-alpha; Cachectin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PBOTNFABI 375.00
Check your price
  • FceR1 alpha Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # A18446
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
IgE Fc receptor subunit alpha; Fc-epsilon RI-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18446 204.00
Check your price
  • SLC15A1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537010
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Peptide transporter 1; Oligopeptide transporter, small intestine isoform; Intestinal H(+)/peptide cotransporter; Proton-coupled dipeptide cotransporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537010 309.00
Check your price
  • Penta-His Tag Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # P21315
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
pentahistidine, Histidine 5X, 5Xhis
Catalog # Size Price (AUD) Your Price (AUD) Qty
P21315 674.00
Check your price
  • Thyroid Stimulating Hormone Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512159
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:200-1:400)
Alternate names:
Thyrotropin alpha chain; Thyroid-stimulating hormone alpha chain; Lutropin alpha chain; Luteinizing hormone alpha chain; Follitropin alpha chain; Follicle-stimulating hormone alpha chain; Chorionic gonadotrophin subunit alpha; Choriogonadotropin alpha chain; Anterior pituitary glycoprotein hormones common subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512159 225.00
Check your price
  • CD147 (soluble) Monoclonal Antibody, Alexa Fluor® 488 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Alexa Fluor® 488
  • Catalog # MA518126
  • Size 100tests
Applications
Flow Cytometry (4ulx100ul whole blood or 10^6 cells)
Alternate names:
Tumor cell-derived collagenase stimulatory factor; OK blood group antigen; Leukocyte activation antigen M6; Extracellular matrix metalloproteinase inducer; Collagenase stimulatory factor; 5F7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518126 275.00
Check your price
  • FSHR Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533548
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (8-15 µg/ml)
Alternate names:
Follitropin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533548 349.00
Check your price
  • Goat anti-Hamster IgG Secondary Antibody, HRP conjugate for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Goat
  • Target Species Hamster
  • Conjugate HRP
  • Catalog # PA129626
  • Size 1mg
Applications
Dot blot (1:15,000-1:65,000), ELISA (1:15,000-1:65,000), Immunohistochemistry (Frozen) (1:2500), Immunohistochemistry (Paraffin) (1:2500), Immunomicroscopy (1:15,000-1:65,000), Western Blot (1:15,000-1:65,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129626 267.00
Check your price
  • STEAP4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520407
  • Size 100ug
Applications
Western Blot (0.5-1 µg/ml)
Alternate names:
Six-transmembrane epithelial antigen of prostate 4; Tumor necrosis factor-alpha-induced adipose-related protein; Dudulin-4; Tumor necrosis factor, alpha-induced protein 9; SixTransMembrane protein of prostate 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520407 349.00
Check your price
  • TIM-1 Monoclonal Antibody, Alexa Fluor® 488 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Alexa Fluor® 488
  • Catalog # MA523634
  • Size 100Tests
Applications
Flow Cytometry (5 µl/10^6 cells)
Alternate names:
T-cell membrane protein 1; T-cell immunoglobulin mucin receptor 1; T-cell immunoglobulin and mucin domain-containing protein 1; Kidney injury molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523634 309.00
Check your price