Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  40  50  60  70  80  90  100  200  300  400  500  600  700  800  900  1000  1630 
211 - 240 of 48881 products displayed
  • RUNX1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSR00271W
  • Size 100µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
SL3/AKV core-binding factor alpha B subunit; SL3-3 enhancer factor 1 alpha B subunit; Polyomavirus enhancer-binding protein 2 alpha B subunit; Oncogene AML-1; Core-binding factor subunit alpha-2; Acute myeloid leukemia 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00271W 375.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor® 488 conjugate for IF, ICC and Flow
  • Host Goat
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 488
  • Catalog # A11034
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11034 209.00
Check your price
  • Phospho-AMPK alpha-1,2 (Thr172) Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 441150G
  • Size 10blots
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (Assay Dependent)
Alternate names:
Hydroxymethylglutaryl-CoA reductase kinase; Acetyl-CoA carboxylase kinase; Tau-protein kinase PRKAA1
Catalog # Size Price (AUD) Your Price (AUD) Qty
441150G 304.00
Check your price
  • Phospho-S6 (Ser244, Ser247) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA125545
  • Size 50µL
Applications
Western Blot (1:1000)
Alternate names:
Phosphoprotein NP33
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125545 349.00
Check your price
  • Cryptosporidium parvum Oocysts Monoclonal Antibody for IF, IHC and ELISA
  • Host Mouse
  • Target Species Protozoa
  • Conjugate Unconjugated
  • Catalog # MA17470
  • Size 100µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50)
Alternate names:
Cryptosporidium parvum, Cryptosporidiosis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17470 299.00
Check your price
  • CD8 alpha Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517048
  • Size 100µL
Applications
Flow Cytometry (1:200-1:400)
Alternate names:
T-lymphocyte differentiation antigen T8/Leu-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517048 225.00
Check your price
  • Goat anti-Mouse IgG2a Cross-Adsorbed Secondary Antibody, Alexa Fluor® 647 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 647
  • Catalog # A21241
  • Size 250µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21241 169.00
Check your price
  • F(ab')2-Goat anti-Rat IgG (H+L) Secondary Antibody, Qdot® 605 conjugate for Western Blot, ICC, IHC and Flow
  • Host Goat
  • Target Species Rat
  • Conjugate Qdot® 605
  • Catalog # Q11601MP
  • Size 200µL
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50), Immunohistochemistry (1:50), Western Blot (1:50)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
Q11601MP 319.00
Check your price
  • Phospho-Tau (Thr181) Monoclonal Antibody for Western Blot, IF and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MN1050
  • Size 100ug
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Paired helical filament-tau; Neurofibrillary tangle protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MN1050 550.00
Check your price
  • Collagen III Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA122147
  • Size 100µL
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (1:4000), Immunohistochemistry (Paraffin) (Assay-dependent), Western Blot (1:100-1:1000)
Alternate names:
EDS4A, FLJ34534
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122147 399.00
Check your price
  • Actin Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Dog, Chicken, Human, Mouse, Non-human primate, Plant, Pig, Protozoa, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA511869
  • Size 500µL
Applications
Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (1:50), Immunoprecipitation (2 ug), Western Blot (1:2500-1:10,000)
Alternate names:
Alpha-actin-1; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511869 309.00
Check your price
  • CD31 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513188
  • Size 500µL
Applications
Flow Cytometry (1:2), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (1:100), Western Blot (1:1-1:100)
Alternate names:
PECA1; GPIIA'; EndoCAM
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513188 309.00
Check your price
  • Cytokeratin 5/6 Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191106
  • Size 250µL
Applications
Immunohistochemistry (Paraffin) (1:25-1:50)
Alternate names:
Type-II keratin Kb5; Keratin-5; Cytokeratin-5; 58 kDa cytokeratin; Type-II keratin Kb6; Keratin-6A; Cytokeratin-6D; Cytokeratin-6A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191106 425.00
Check your price
  • GLUT1 Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA137783
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
HepG2 glucose transporter; Glucose transporter type 1, erythrocyte/brain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA137783 475.00
Check your price
  • IL-1 beta Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 701304
  • Size 100µg
Applications
Western Blot (2-3 ug/ml)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
701304 303.00
Check your price
  • Borrelia burgdorferi Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA121685
  • Size 500µg
Applications
ELISA (Assay dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Western Blot (Assay dependent)
Alternate names:
B. burgdorferi, Lyme disease, Borrelia
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121685 349.00
Check your price
  • Donkey anti-Sheep IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 680 conjugate for Western Blot, IF and ICC
  • Host Donkey
  • Target Species Sheep
  • Conjugate Alexa Fluor® 680
  • Catalog # A21102
  • Size 500µL
Applications
Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL), Western Blot (0.04-0.2 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21102 209.00
Check your price
  • Estrogen Receptor alpha Polyclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA1308
  • Size 50µg
Applications
Immunocytochemistry (1:1000-1:4000), Immunofluorescence (1:1000-1:4000), Immunohistochemistry (Frozen) (1-5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1308 349.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor® 555 conjugate for IF, ICC and Flow
  • Host Goat
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 555
  • Catalog # A21429
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21429 209.00
Check your price
  • Normal Rabbit Serum for Ctrl
  • Host Rabbit
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # 016101
  • Size 10mL
Applications
Control (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
016101 39.00
Check your price
  • AP2 gamma Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517330
  • Size 100µL
Applications
Immunocytochemistry (1:400), Immunofluorescence (1:400), Western Blot (1:1000)
Alternate names:
Transcription factor ERF-1; Activating enhancer-binding protein 2 gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517330 275.00
Check your price
  • Vaccinia Virus Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA174226
  • Size 200µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA174226 399.00
Check your price
  • Chicken anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 647 conjugate for IF, ICC and IHC
  • Host Chicken
  • Target Species Rat
  • Conjugate Alexa Fluor® 647
  • Catalog # A21472
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21472 169.00
Check your price
  • Phospho-CaMKII alpha/beta/delta (Thr305) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538238
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Calcium/calmodulin-dependent protein kinase type II subunit alpha, CaM kinase II subunit alpha, CaMK-II subunit alpha, CAMK2A, CAMKA, KIAA0968, CaMK2-a, CaMK2a, CaMK-2a, Calcium/calmodulin-dependent protein kinase type II subunit delta, CaM kinase II subunit delta, CaMK-II subunit delta, CAMK2D, CAMKD, CaMK2- delta, CaMK2 delta, CaMK-2 delta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538238 309.00
Check your price
  • VEGF Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA516754
  • Size 500µL
Applications
Immunocytochemistry (1:40-1:200), Immunofluorescence (1:40-1:200), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (3 µg/ml), Western Blot (1 µg/mL)
Alternate names:
Vascular permeability factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516754 309.00
Check your price
  • hCG beta-2 Monoclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516741
  • Size 500ug
Applications
ELISA (1:100-1:500), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516741 349.00
Check your price
  • Macrophages / Monocytes Monoclonal Antibody for IHC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Bovine, Dog, Deer, Horse, Cat, Guinea pig, Goat, Human, Mammal, Non-human primate, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA180446
  • Size 200µg
Applications
Flow Cytometry (1/50-1/100), Immunohistochemistry (Paraffin, Frozen) (1/100-1/200)
Alternate names:
60B8AG, CAGB, CFAG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14, NIF, P14
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180446 399.00
Check your price
  • NFkB p65 Polyclonal Antibody for Western Blot, IF, ICC, Flow, IP and ChIP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1186
  • Size 100ug
Applications
ChIP assay (1-5 µg x 10^6 cells), Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunoprecipitation (3 µg), Western Blot (1-3 µg/ml)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; Nuclear factor NF-kappa-B p65 subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1186 309.00
Check your price
  • NOXA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA118332
  • Size 100µL
Applications
Western Blot (1:1,000)
Alternate names:
Protein Noxa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118332 349.00
Check your price
  • Laminin alpha-3 Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524246
  • Size 100µg
Applications
Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (2 µg/ml)
Alternate names:
Nicein subunit alpha; Laminin-7 subunit alpha; Laminin-6 subunit alpha; Laminin-5 subunit alpha; Kalinin subunit alpha; Epiligrin subunit alpha; Epiligrin 170 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524246 275.00
Check your price