Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  50  60  66  67  68  69  70  71  72  73  74  75  76  77  78  79  80  81  82  83  84  85  86  87  88  89  90  91  92  93  94  95  100  110  120  130  140  150  160  170  180  200  300  400  500  600  700  800  900  1000  1630 
2401 - 2430 of 48882 products displayed
  • Goat anti-Human IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate HRP
  • Catalog # A18805
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18805 109.00
Check your price
  • CD204 Polyclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA522956
  • Size 100µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Frozen) (Assay-Dependent), Western Blot (0.5 µg/ml)
Alternate names:
Scavenger receptor type A; Macrophage acetylated LDL receptor I and II; Scavenger receptor class A member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522956 349.00
Check your price
  • beta Tubulin Polyclonal Antibody for Western Blot, IHC (P), IP and FN
  • Host Rabbit
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # PA121153
  • Size 500µL
Applications
Functional Assay (Assay-dependent), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Tubulin beta-5 chain; Tubulin beta-4 chain; Tubulin 5 beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121153 390.00
Check your price
  • SIAH1 Polyclonal Antibody for Western Blot, IP and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517998
  • Size 100µg
Applications
Immunoprecipitation (Assay dependent), Western Blot (1-3 µg/ml)
Alternate names:
Siah-1a; Seven in absentia homolog 1; Seven in absentia homolog 1a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517998 349.00
Check your price
  • DMC1 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526337
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
DMC1H, HsLim15, LIM15, dJ199H16.1, DMC1 homologue, disrupted meiotic cDNA1, yeast, homolog of, meiotic recombination protein DMC1/LIM15 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526337 349.00
Check your price
  • FOXQ1 Monoclonal Antibody for IF, ICC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA522893
  • Size 100µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml)
Alternate names:
Hepatocyte nuclear factor 3 forkhead homolog 1; HNF-3/forkhead-like protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA522893 309.00
Check your price
  • CA150 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA540604
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Transcription factor CA150; TATA box-binding protein-associated factor 2S; p144; Formin-binding protein 28
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540604 309.00
Check your price
  • ADAMDEC1 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526128
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
A disintegrin and metalloproteinase domain-like protein decysin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526128 349.00
Check your price
  • IRF4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517254
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Transcriptional activator PIP; PU.1 interaction partner; NF-EM5; Lymphocyte-specific interferon regulatory factor; Multiple myeloma oncogene 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517254 275.00
Check your price
  • ErbB3 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA512675
  • Size 500µL
Applications
Western Blot (4 µg/mL)
Alternate names:
Tyrosine kinase-type cell surface receptor HER3; Proto-oncogene-like protein c-ErbB-3; Glial growth factor receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512675 349.00
Check your price
  • Phospho-14-3-3 beta (Ser58) Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA14612
  • Size 100µL
Applications
Immunohistochemistry (1:500), Western Blot (1:1000)
Alternate names:
Protein kinase C inhibitor protein 1; Protein 1054; Prepronerve growth factor RNH-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14612 375.00
Check your price
  • Ku70 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA123300
  • Size 100µL
Applications
Western Blot (1:2500)
Alternate names:
X-ray repair complementing defective repair in Chinese hamster cells 6; Thyroid-lupus autoantigen; Lupus Ku autoantigen protein p70; DNA repair protein XRCC6; CTC box-binding factor 75 kDa subunit; ATP-dependent DNA helicase II 70 kDa subunit; ATP-dependent DNA helicase 2 subunit 1; 70 kDa subunit of Ku antigen; 5'-deoxyribose-5-phosphate lyase Ku70
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123300 309.00
Check your price
  • Phalloidin, DyLight® 488 conjugate for IF, ICC and IHC
  • Host
  • Target Species Many
  • Conjugate DyLight® 488
  • Catalog # 21833
  • Size 300units
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Assay dependent)
Alternate names:
DY488-Phalloidin, F-actin toxin, Amanita phalloides
Catalog # Size Price (AUD) Your Price (AUD) Qty
21833 374.00
Check your price
  • GSK3 beta Monoclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515109
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Serine/threonine-protein kinase GSK3B; Factor A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515109 275.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, TRITC conjugate for IF
  • Host Goat
  • Target Species Chicken
  • Conjugate TRITC
  • Catalog # A16059
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16059 94.00
Check your price
  • CD69 Monoclonal Antibody, PE-Cyanine7 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE-Cyanine7
  • Catalog # A16361
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
MLR-3; Leukocyte surface antigen Leu-23; GP32/28; Early T-cell activation antigen p60; EA1; C-type lectin domain family 2 member C; BL-AC/P26; Activation inducer molecule
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16361 151.00
Check your price
  • HAUS4 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA525153
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
C14orf94, HAUS augmin-like complex subunit 4, homologous to Augmin subunits
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525153 349.00
Check your price
  • NKPD1 Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA524399
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
NTPase KAP family P-loop domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524399 349.00
Check your price
  • CD4 Monoclonal Antibody, Biotin conjugate for IHC (F), IHC (P), Flow and IP
  • Host Mouse
  • Target Species Rat
  • Conjugate Biotin
  • Catalog # MA517389
  • Size 500ug
Applications
Flow Cytometry (0.5 ug/10^6 cells), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
W3/25 antigen; T-cell surface antigen T4/Leu-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517389 309.00
Check your price
  • Rabbit anti-Mouse IgG Fc Secondary Antibody, AP conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Mouse
  • Conjugate AP
  • Catalog # 31332
  • Size 1mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:50,000), Immunofluorescence (1:5,000-1:50,000), Immunohistochemistry (1:5,000-1:50,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:50,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31332 194.00
Check your price
  • MMP9 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513199
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Gelatinase B; 92 kDa type IV collagenase; 92 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513199 349.00
Check your price
  • TIA-1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529685
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
p40-TIA-1; T-cell-restricted intracellular antigen-1; RNA-binding protein TIA-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529685 349.00
Check your price
  • CD3d Monoclonal Antibody, PE-Cyanine5.5 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE-Cyanine5.5
  • Catalog # MHCD0318
  • Size 0.5ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
T-cell receptor T3 delta chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD0318 319.00
Check your price
  • Trypsin Monoclonal Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # HYB02103B01
  • Size 100µl
Applications
ELISA (1:16,000), Western Blot (Assay dependent)
Alternate names:
Trypsin I; Serine protease 1; Cationic trypsinogen; Beta-trypsin
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB02103B01 700.00
Check your price
  • Goat anti-Human Kappa Light Chain Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate AP
  • Catalog # A18856
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18856 220.00
Check your price
  • IkB alpha Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA141127
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
Major histocompatibility complex enhancer-binding protein MAD3; I-kappa-B-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141127 309.00
Check your price
  • Desmocollin 3 Polyclonal Antibody for Western Blot and IHC (F)
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547909
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
ARVC10, ARVD10, CDHF5, CMD1BB, HDGC, HDGC, cadherin family member 5, desmoglein-2, CDHF6, PVA, 130 kDa pemphigus vulgaris antigen, 130-kD pemphigus vulgaris antigen, cadherin family member 6, desmoglein-3, pemphigus vulgaris antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547909 375.00
Check your price
  • CRY1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513124
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Cryptochrome-1; PHLL1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513124 349.00
Check your price
  • CDK2 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516346
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (1:100)
Alternate names:
p33 protein kinase; Cell division protein kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516346 499.00
Check your price
  • UBE2L6 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA542284
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Ubiquitin-protein ligase L6; Ubiquitin carrier protein L6; E2 ubiquitin-conjugating enzyme L6; UbcM8; UbcH8; Retinoic acid-induced gene B protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542284 309.00
Check your price