Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  50  60  70  78  79  80  81  82  83  84  85  86  87  88  89  90  91  92  93  94  95  96  97  98  99  100  101  102  103  104  105  106  107  110  120  130  140  150  160  170  180  190  200  300  400  500  600  700  800  900  1000  1630 
2761 - 2790 of 48882 products displayed
  • MCP-2 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523732
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (1-4 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Small-inducible cytokine A8; Monocyte chemotactic protein 2; Monocyte chemoattractant protein 2; HC14
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523732 375.00
Check your price
  • GEM Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521298
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
RAS-like protein KIR; GTP-binding mitogen-induced T-cell protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521298 349.00
Check your price
  • Claudin 2 Polyclonal Antibody for Western Blot, IF and ELISA
  • Host Rabbit
  • Target Species Dog, Human, Rat
  • Conjugate Unconjugated
  • Catalog # 516100
  • Size 100µg
Applications
ELISA (0.1-1.0 ug/ml), Immunofluorescence (1-3 ug/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
SP82
Catalog # Size Price (AUD) Your Price (AUD) Qty
516100 303.00
Check your price
  • OLIG2 Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523456
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (1:200-1:400)
Alternate names:
Protein kinase C-binding protein RACK17; Protein kinase C-binding protein 2; Class E basic helix-loop-helix protein 19; Class B basic helix-loop-helix protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523456 349.00
Check your price
  • Phospho-CDK9 (Thr186) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517197
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Cell division protein kinase 9; Tat-associated kinase complex catalytic subunit; Serine/threonine-protein kinase PITALRE; Cell division cycle 2-like protein kinase 4; C-2K
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517197 309.00
Check your price
  • GSC Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA540495
  • Size 100µl
Applications
Immunohistochemistry (5 µg/ml), Western Blot (0.2-1 µg/ml )
Alternate names:
Goosecoid homeobox
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540495 309.00
Check your price
  • GLUT1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Bovine, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA11063
  • Size 100µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay dependent), Western Blot (1:1000)
Alternate names:
Glucose transporter type 1, erythrocyte/brain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11063 349.00
Check your price
  • SP4 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA541444
  • Size 100µl
Applications
Immunohistochemistry (5 µg/ml), Western Blot (0.2-1 µg/ml)
Alternate names:
SPR-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541444 309.00
Check your price
  • Endostatin Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA524199
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
KNO, KNO1, KS, antiangiogenic agent, collagen alpha-1(XVIII) chain, endostatin, multi-functional protein MFP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524199 349.00
Check your price
  • ZO-1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA528858
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Zonula occludens protein 1; Zona occludens protein 1; Tight junction protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528858 349.00
Check your price
  • Rabbit anti-Goat IgG (H+L) Secondary Antibody for ELISA
  • Host Rabbit
  • Target Species Goat
  • Conjugate Unconjugated
  • Catalog # A16138
  • Size 2mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16138 106.00
Check your price
  • ISLR Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524804
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
HsT17563, immunoglobulin superfamily containing leucine-rich repeat protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524804 349.00
Check your price
  • ZNF306 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA522646
  • Size 100µg
Applications
ELISA (Assay-Dependent), Western Blot (1-5 µg/ml)
Alternate names:
Zinc finger protein 47 homolog; Zinc finger protein 309; Zinc finger protein 306; Zinc finger and SCAN domain-containing protein 13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA522646 309.00
Check your price
  • Canine Distemper Virus Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA122748
  • Size 100µg
Applications
ELISA (Assay dependent)
Alternate names:
CDV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122748 309.00
Check your price
  • 14-3-3 beta Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFPA0077
  • Size 100µL
Applications
Western Blot (1:2000)
Alternate names:
Protein kinase C inhibitor protein 1; Protein 1054; Prepronerve growth factor RNH-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFPA0077 309.00
Check your price
  • DLAT Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529043
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:5000-1:20,000)
Alternate names:
Pyruvate dehydrogenase complex component E2; M2 antigen complex 70 kDa subunit; Dihydrolipoamide acetyltransferase component of pyruvate dehydrogenase complex; 70 kDa mitochondrial autoantigen of primary biliary cirrhosis
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529043 349.00
Check your price
  • ITGB5 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527759
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
integrin beta-5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527759 349.00
Check your price
  • Goat anti-Mouse IgG Fc Secondary Antibody, HRP conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # 31437
  • Size 2mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:100,000), Immunofluorescence (1:5,000-1:100,000), Immunohistochemistry (1:5,000-1:100,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:200,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31437 199.00
Check your price
  • S100A10 Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524080
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
p11; p10 protein; S100 calcium-binding protein A10; Cellular ligand of annexin II; Calpactin-1 light chain; Calpactin I light chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524080 275.00
Check your price
  • ICAM-2 Monoclonal Antibody for Flow, IP and FN
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA516837
  • Size 500ug
Applications
Flow Cytometry (1:50-1:200), Functional Assay (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
Lymphocyte function-associated AG-1 counter-receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516837 499.00
Check your price
  • PKA alpha Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517626
  • Size 100µL
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
PKA C-alpha; cAMP-dependent protein kinase catalytic subunit alpha; cAMP-dependent protein kinase catalytic subunit alpha, isoform 1; protein kinase A catalytic subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517626 275.00
Check your price
  • ZIC1/ZIC2/ZIC3 Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536307
  • Size 100µl
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Zinc finger protein of the cerebellum 3; Zinc finger protein of the cerebellum 1; Zinc finger protein of the cerebellum 2; Zinc finger protein 201; Zinc finger protein 203
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536307 309.00
Check your price
  • FAK Polyclonal Antibody for Western Blot and IF
  • Host Rabbit
  • Target Species Amphibian, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA516676
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Western Blot (5-10 µg/mL)
Alternate names:
pp125FAK; p125FAK; Protein-tyrosine kinase 2; Focal adhesion kinase-related nonkinase; Protein phosphatase 1 regulatory subunit 71
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516676 309.00
Check your price
  • Donkey anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Donkey
  • Target Species Goat
  • Conjugate Biotin
  • Catalog # A16009
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16009 136.00
Check your price
  • NMDAR1 Polyclonal Antibody for Western Blot and IHC (F)
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OPA104011
  • Size 25µg
Applications
Immunohistochemistry (Frozen) (1:1000 to 1:2000), Western Blot (1:2,000)
Alternate names:
N-methyl-D-aspartate receptor subunit NR1; Glutamate [NMDA] receptor subunit zeta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
OPA104011 375.00
Check your price
  • Phospho-eIF2a (Ser51) Oligoclonal Antibody for Western Blot, IF, ICC and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710292
  • Size 100µg
Applications
ChIP assay (1 ul), Immunocytochemistry (1-3 ug/ml), Immunofluorescence (1-3 ug/ml), Western Blot (2-3 ug/ml)
Alternate names:
Eukaryotic translation initiation factor 2 subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
710292 303.00
Check your price
  • S6 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Fruit fly, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515123
  • Size 100µL
Applications
Flow Cytometry (1:25), Immunocytochemistry (1:25), Immunofluorescence (1:25), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
Phosphoprotein NP33
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515123 275.00
Check your price
  • CD34 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate PE
  • Catalog # MA110206
  • Size 100Tests
Applications
Flow Cytometry (20 ul/10^6 cells)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110206 275.00
Check your price
  • Acetyl-NFkB p65 (Lys310) Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517264
  • Size 100µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; Nuclear factor NF-kappa-B p65 subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517264 309.00
Check your price
  • PDK2 Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA535376
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (1:1000)
Alternate names:
Pyruvate dehydrogenase kinase isoform 2; PDK P45
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535376 349.00
Check your price