Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  50  60  70  82  83  84  85  86  87  88  89  90  91  92  93  94  95  96  97  98  99  100  101  102  103  104  105  106  107  108  109  110  111  120  130  140  150  160  170  180  190  200  300  400  500  600  700  800  900  1000  1630 
2881 - 2910 of 48877 products displayed
  • Cystatin C Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA528702
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Cystatin-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528702 349.00
Check your price
  • HTRA2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520064
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 µg/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Serine proteinase OMI; Serine protease 25; Omi stress-regulated endoprotease; High temperature requirement protein A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520064 349.00
Check your price
  • Phospho-Tau (Ser396) Oligoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 710298
  • Size 100µg
Applications
ELISA (Assay-Dependent), Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (1:500-1:5000)
Alternate names:
Paired helical filament-tau; Neurofibrillary tangle protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
710298 305.00
Check your price
  • TCIRG1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534419
  • Size 100ug
Applications
Western Blot (0.5 ug/mL)
Alternate names:
Vacuolar proton translocating ATPase 116 kDa subunit a isoform 3; T-cell immune response cDNA7 protein; T-cell immune regulator 1; Osteoclastic proton pump 116 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534419 349.00
Check your price
  • SUFU Monoclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA514954
  • Size 100µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
OTTHUMP00000020377; OTTHUMP00000020378; OTTHUMP00000020379; suppressor of fused homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514954 275.00
Check your price
  • Glutamate Dehydrogenase Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529492
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
GLUD2, GLUD1, GDH2, GLUDP1, GDH 2, glutamate dehydrogenase 2, mitochondrial, glutamate dehydrogenase pseudogene 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529492 349.00
Check your price
  • CD41 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human, Mustelid, Mink, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA516920
  • Size 1mg
Applications
Flow Cytometry (1:100 -1:200)
Alternate names:
Platelet membrane glycoprotein IIb; GPalpha IIb
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516920 800.00
Check your price
  • FAS Monoclonal Antibody for Flow and FN
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119024
  • Size 100µg
Applications
Flow Cytometry (2-10 ug/ml), Functional Assay (Assay-Dependent)
Alternate names:
FASLG receptor; Apoptosis-mediating surface antigen FAS; Apo-1 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119024 225.00
Check your price
  • Cardiac Troponin T Monoclonal Antibody for IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Avian, Dog, Chicken, Fish, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA512960
  • Size 500µL
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunomicroscopy (Assay Dependent)
Alternate names:
Cardiac muscle troponin T
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512960 309.00
Check your price
  • AXIN2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521093
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20µg/ml), Immunohistochemistry (Paraffin) (5 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Conductin; Axis inhibition protein 2; Axin-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521093 349.00
Check your price
  • GPR18 Polyclonal Antibody for IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532809
  • Size 50ug
Applications
ELISA (Assay-dependent), Immunohistochemistry (Paraffin) (5 µg/ml)
Alternate names:
G-protein coupled receptor 18
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532809 349.00
Check your price
  • CD11b Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # RM2801
  • Size 1mL
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
CD11B, CR3A, MAC-1, MAC1A, MGC117044, MO1A, SLEB6, Integrin alpha M
Catalog # Size Price (AUD) Your Price (AUD) Qty
RM2801 117.00
Check your price
  • Ovalbumin Polyclonal Antibody, HRP conjugate for Western Blot and ELISA
  • Host Rabbit
  • Target Species Chicken
  • Conjugate HRP
  • Catalog # PA1196HRP
  • Size 100µg
Applications
ELISA (1:1000-1:5000), Western Blot (1:1000-1:5000)
Alternate names:
Plakalbumin; Egg albumin; Allergen Gal d II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1196HRP 225.00
Check your price
  • MBP Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA516413
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Myelin membrane encephalitogenic protein; Myelin A1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516413 475.00
Check your price
  • SLC39A7 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521072
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Zrt-, Irt-like protein 7; Solute carrier family 39 member 7; Really interesting new gene 5 protein; Histidine-rich membrane protein Ke4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521072 349.00
Check your price
  • Granzyme A Monoclonal Antibody for IF, ICC, Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524105
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Immunoprecipitation (25 µg/ml)
Alternate names:
Hanukkah factor; Granzyme-1; Fragmentin-1; Cytotoxic T-lymphocyte proteinase 1; CTL tryptase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524105 275.00
Check your price
  • EpCAM Monoclonal Antibody for IF, ICC, IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512153
  • Size 500µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (1-2 µg/ml)
Alternate names:
Tumor-associated calcium signal transducer 1; Major gastrointestinal tumor-associated protein GA733-2; KSA; KS 1/4 antigen; Epithelial glycoprotein 314; Epithelial glycoprotein; Epithelial cell surface antigen; Cell surface glycoprotein Trop-1; Adenocarcinoma-associated antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512153 309.00
Check your price
  • K-cadherin Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547373
  • Size 100µg
Applications
Flow Cytometry (0.25 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Kidney cadherin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547373 349.00
Check your price
  • TET2 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518299
  • Size 100µL
Applications
Immunoprecipitation (1:100-1:500), Western Blot (1:500-1:3000)
Alternate names:
KIAA1546, MDS, tet methylcytosine dioxygenase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518299 309.00
Check your price
  • Pancreatic Amylase Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110619
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
1,4-alpha-D-glucan glucanohydrolase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110619 399.00
Check your price
  • Gpr143 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # OSR00147W
  • Size 100µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Ocular albinism type 1 protein homolog; MOA1
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00147W 309.00
Check your price
  • Phospho-CXCR4 (Ser339) Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538461
  • Size 100µg
Applications
Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Stromal cell-derived factor 1 receptor; NPYRL; Lipopolysaccharide-associated protein 3; Leukocyte-derived seven transmembrane domain receptor; LCR1; HM89; Fusin; FB22; Pre-B-cell-derived chemokine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538461 309.00
Check your price
  • BID Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517034
  • Size 100µL
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
p22 BID
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517034 309.00
Check your price
  • LTK Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA141118
  • Size 100µg
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Protein tyrosine kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141118 309.00
Check your price
  • Vanilloid Receptor 1 (residues 7-21) Synthetic Peptide for Ctrl
  • Host N/A
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # PEP202
  • Size 50µg
Applications
Control (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PEP202 98.00
Check your price
  • PBXIP1 Polyclonal Antibody for IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535941
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000)
Alternate names:
Hematopoietic PBX-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535941 309.00
Check your price
  • Phospho-WASP (Tyr290) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538346
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
IMD2, SCNX, THC, THC1, Thrombocytopenia 1 (X linked), U42471, Was, WASp, WASP, Wiskott Aldrich syndrome (eczema thrombocytopenia), Wiskott Aldrichsyndrome protein, Wiskott-Aldrich syndrome protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538346 309.00
Check your price
  • TREM1 Polyclonal Antibody for ELISA
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA175482
  • Size 1mL
Applications
ELISA (Assay Dependent)
Alternate names:
Triggering receptor expressed on monocytes 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175482 425.00
Check your price
  • PYGO2 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524240
  • Size 100µg
Applications
Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Western Blot (0.5-5 µg/ml)
Alternate names:
PP7910, 1190004M21Rik, pygopus homolog 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524240 275.00
Check your price
  • IL-31 Polyclonal Antibody for Western Blot, IF, ICC, ELISA and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547405
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Neutralization (1 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
interleukin-31
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547405 349.00
Check your price