Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  30  40  50  60  70  80  90  100  110  117  118  119  120  121  122  123  124  125  126  127  128  129  130  131  132  133  134  135  136  137  138  139  140  141  142  143  144  145  146  150  160  170  180  190  200  210  220  230  300  400  500  600  700  800  900  1000  1100  1630 
3931 - 3960 of 48882 products displayed
  • hnRNP M Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA134962
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (3:25), Western Blot (1:1000)
Alternate names:
M4 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134962 399.00
Check your price
  • HMGCR Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537367
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
3 hydroxy 3 methylglutaryl Coenzyme A reductase, 3 hydroxymethylglutaryl CoA reductase, 3-hydroxy-3-methylglutaryl-coenzyme A reductase, 3H3M, HMDH, HMG CoA reductase, HMG CoAR, HMG-CoA reductase, Hmgcr, Hydroxymethylglutaryl CoA reductase, LDLCQ3, MGC103269
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537367 399.00
Check your price
  • HMBS Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537366
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
Pre-uroporphyrinogen synthase; Hydroxymethylbilane synthase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537366 399.00
Check your price
  • HLA-DRB3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537365
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
MHC class II antigen DRB3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537365 399.00
Check your price
  • HFE Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537364
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
HLA-H
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537364 399.00
Check your price
  • HDAC3 Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA518069
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:400-1:800)
Alternate names:
SMAP45; RPD3-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518069 399.00
Check your price
  • GTF2H1 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518316
  • Size 100µl
Applications
Immunoprecipitation (1:50-1:200), Western Blot (1:300-1:1000)
Alternate names:
TFIIH basal transcription factor complex p62 subunit; General transcription factor IIH polypeptide 1; Basic transcription factor 2 62 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518316 399.00
Check your price
  • GSS Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537307
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
Glutathione synthase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537307 399.00
Check your price
  • GPX5 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537309
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
Glutathione peroxidase 5; Epididymis-specific glutathione peroxidase-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537309 399.00
Check your price
  • GPX1 Polyclonal Antibody for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Sheep
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA118279
  • Size 100µL
Applications
ELISA (1:2,000-1:20,000), Immunohistochemistry (Paraffin, Frozen) (1:2,000-1:20,000), Western Blot (1:10,000-1:100,000)
Alternate names:
Cellular glutathione peroxidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118279 399.00
Check your price
  • Golgi protein 58k Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IM
  • Host Mouse
  • Target Species Bovine, Dog, Hamster, Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA122144
  • Size 100µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:250), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay dependent), Western Blot (1:5000)
Alternate names:
LCHC1; Formiminotransferase-cyclodeaminase; 58 kDa microtubule-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122144 399.00
Check your price
  • GIPC2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537334
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
SemaF cytoplasmic domain-associated protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537334 399.00
Check your price
  • GAS2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537317
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
GAS 2, GAS-2, Gas2, GAS2, Growth arrest specific 2, Growth arrest specific protein 2, Growth arrest-specific protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537317 399.00
Check your price
  • gamma Adaptin Polyclonal Antibody for Western Blot, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA118306
  • Size 100µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1:200-1:2,000), Western Blot (1:1,000-1:4,000)
Alternate names:
Golgi adaptor HA1/AP1 adaptin subunit gamma-1; Gamma1-adaptin; Clathrin assembly protein complex 1 gamma-1 large chain; Adaptor-related protein complex 1 subunit gamma-1; Adaptor protein complex AP-1 subunit gamma-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118306 399.00
Check your price
  • FRS2 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA124685
  • Size 100µL
Applications
Immunoprecipitation (10-20 µg per 3 mg RIPA), Western Blot (1:1000)
Alternate names:
Suc1-associated neurotrophic factor target 1; FGFR-signaling adaptor SNT; FRS2-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124685 399.00
Check your price
  • FAM3B Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537311
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
Pancreatic-derived factor; Cytokine-like protein 2-21
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537311 399.00
Check your price
  • eIF6 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537371
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
p27(BBP); CAB; B4 integrin interactor; B(2)GCN homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537371 399.00
Check your price
  • EAPP Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA518062
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:200)
Alternate names:
E2F-associated phosphoprotein; C14orf11; BM-036
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518062 399.00
Check your price
  • E2F3 Monoclonal Antibody for Western Blot, IP and GS
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA125333
  • Size 100µL
Applications
Gel Shift (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (4-8 µg/ml)
Alternate names:
RP1-177P22.2, DKFZp686C18211, E2F-3, KIAA0075, MGC104598
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125333 399.00
Check your price
  • Dynactin 1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537360
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
p150-glued; p135; DAP-150; 150 kDa dynein-associated polypeptide
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537360 399.00
Check your price
  • Dynactin 1 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518308
  • Size 100µl
Applications
Immunoprecipitation (1:50-1:200), Western Blot (1:1000)
Alternate names:
p150-glued; p135; DAP-150; 150 kDa dynein-associated polypeptide
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518308 399.00
Check your price
  • DsbA Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA518312
  • Size 100µl
Applications
Western Blot (1:1000)
Alternate names:
B21_03695, ECD_03746, dsbA, Thiol:disulfide interchange protein, Disulphide oxidoreductase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518312 399.00
Check your price
  • DP2 Monoclonal Antibody for Western Blot, IP and GS
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191207
  • Size 100µL
Applications
Gel Shift (Assay-dependent), Immunoprecipitation (Assay dependent), Western Blot (0.5-1 µg/ml)
Alternate names:
E2F dimerization partner 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191207 399.00
Check your price
  • Dopamine beta Hydroxylase Polyclonal Antibody for Western Blot, IHC, RIA and IA
  • Host Rabbit
  • Target Species Bovine, Guinea pig, Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA118314
  • Size 100µL
Applications
Immunohistochemistry (1:500-1:2000), Inhibition Assays (1:500-1:2000), Radioimmune Assays (1:500-1:2000), Western Blot (1:500-1:2000)
Alternate names:
Dopamine beta-monooxygenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118314 399.00
Check your price
  • DNAI1 Monoclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Chicken, Deer, Hamster, Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA122146
  • Size 100µL
Applications
ELISA (Assay dependent), Immunocytochemistry (Assay dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:2000)
Alternate names:
Axonemal dynein intermediate chain 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122146 399.00
Check your price
  • DNAH9 Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Chicken, Human
  • Conjugate Unconjugated
  • Catalog # MA126770
  • Size 100µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent), Western Blot (1:100)
Alternate names:
Ciliary dynein heavy chain 9; Axonemal beta dynein heavy chain 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126770 399.00
Check your price
  • Dipyrone Polyclonal Antibody for ELISA
  • Host Sheep
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # PA175123
  • Size 100µL
Applications
ELISA (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175123 399.00
Check your price
  • DDB2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537361
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
Damage-specific DNA-binding protein 2; UV-damaged DNA-binding protein 2; DDB p48 subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537361 399.00
Check your price
  • CYP11A1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537359
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
Cytochrome P450(scc); Cytochrome P450 11A1; Cholesterol desmolase; CYPXIA1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537359 399.00
Check your price
  • CYBA Monoclonal Antibody for Western Blot, IHC (F), Flow, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518047
  • Size 100µL
Applications
ELISA (Assay-dependent), Flow Cytometry (Assay-dependent), Immunohistochemistry (Frozen) (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:1000)
Alternate names:
p22-phox; p22 phagocyte B-cytochrome; Superoxide-generating NADPH oxidase light chain subunit; Neutrophil cytochrome b 22 kDa polypeptide; Cytochrome b558 subunit alpha; Cytochrome b(558) alpha chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518047 399.00
Check your price