Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  50  60  70  80  90  100  110  120  130  134  135  136  137  138  139  140  141  142  143  144  145  146  147  148  149  150  151  152  153  154  155  156  157  158  159  160  161  162  163  170  180  190  200  210  220  230  240  300  400  500  600  700  800  900  1000  1100  1630 
4441 - 4470 of 48882 products displayed
  • Laminin beta-1 Monoclonal Antibody, Biotin conjugate for Western Blot, IF, IHC (F), IHC (P) and IP
  • Host Rat
  • Target Species Hamster, Human, Mouse, Pig
  • Conjugate Biotin
  • Catalog # MA514654
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (2 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Laminin-8 subunit beta; Laminin-6 subunit beta; Laminin-2 subunit beta; Laminin-12 subunit beta; Laminin-10 subunit beta; Laminin-1 subunit beta; Laminin B1 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514654 375.00
Check your price
  • Fibronectin Polyclonal Antibody for IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA123693
  • Size 500µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1:100)
Alternate names:
Cold-insoluble globulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123693 375.00
Check your price
  • Cyclin B1 Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA514324
  • Size 500µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (2-4 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1:100-1:1000)
Alternate names:
G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1,OTTHUMP00000223023
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514324 375.00
Check your price
  • Calcitonin Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516325
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Calcitonin gene-related peptide I; Alpha-type CGRP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516325 375.00
Check your price
  • Adenovirus Type 2 Fiber Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA134961
  • Size 500µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Western Blot (1:200)
Alternate names:
HADV, AdV, Adeno Virus, HADV-2, AdV-2, Adeno Virus 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134961 375.00
Check your price
  • Resistin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523784
  • Size 500µg
Applications
ELISA (0.5-2 µg/ml (Detection), 2-8 µg/ml (Capture))
Alternate names:
Cysteine-rich secreted protein FIZZ3; Cysteine-rich secreted protein A12-alpha-like 2; C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein; Adipose tissue-specific secretory factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523784 375.00
Check your price
  • Plasminogen Monoclonal Antibody for IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120080
  • Size 500µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
RP1-81D8.1, DKFZp779M0222
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120080 375.00
Check your price
  • Neutrophil elastase Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110606
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
PMN elastase; Medullasin; Human leukocyte elastase; Elastase-2; Bone marrow serine protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110606 375.00
Check your price
  • MCP-2 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523732
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (1-4 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Small-inducible cytokine A8; Monocyte chemotactic protein 2; Monocyte chemoattractant protein 2; HC14
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523732 375.00
Check your price
  • Lidocaine Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA110751
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
Lignocaine
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110751 375.00
Check your price
  • IL-10 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523796
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.005-0.015 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Cytokine synthesis inhibitory factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523796 375.00
Check your price
  • FABP4 Polyclonal Antibody for Western Blot and ELISA
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA121216
  • Size 500µg
Applications
ELISA (1:1000-1:5000), Western Blot (1:1000-1:5000)
Alternate names:
Fatty acid-binding protein 4; Adipocyte-type fatty acid-binding protein; Adipocyte lipid-binding protein; Protein 422; P2 adipocyte protein; P15; Myelin P2 protein homolog; 3T3-L1 lipid-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121216 375.00
Check your price
  • EGF Monoclonal Antibody for IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523704
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Immunohistochemistry (8-25 µg/ml)
Alternate names:
Epidermal Growth Factor, URG, Urogastrone, HOMG4, URG, beta-urogastrone, pro-epidermal growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523704 375.00
Check your price
  • CXCL8 Monoclonal Antibody for Western Blot, IF, ICC, Flow, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523697
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Neutralization (Assay-Dependent), Western Blot (1 µg/ml)
Alternate names:
T-cell chemotactic factor; Protein 3-10C; Neutrophil-activating protein 1; Monocyte-derived neutrophil-activating peptide; Monocyte-derived neutrophil chemotactic factor; Granulocyte chemotactic protein 1; Emoctakin; Chemokine (C-X-C motif) ligand 8; C-X-C motif chemokine 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523697 375.00
Check your price
  • CXCL10 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523774
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (10-40 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Small-inducible cytokine B10; Interferon-gamma induced protein CRG-2; C7; 10 kDa interferon gamma-induced protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523774 375.00
Check your price
  • CXCL1 Monoclonal Antibody for Western Blot, Flow, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523707
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Neutralization (3-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Neutrophil-activating protein 3; Melanoma growth stimulatory activity; GRO-alpha(1-73); C-X-C motif chemokine 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523707 375.00
Check your price
  • Chymotrypsin Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110623
  • Size 500µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Caldecrin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110623 375.00
Check your price
  • CA125 Monoclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA124603
  • Size 500µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Ovarian carcinoma antigen CA125; Ovarian cancer-related tumor marker CA125
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124603 375.00
Check your price
  • Cyclophilin A Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Hamster, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1025
  • Size 400µL
Applications
Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Western Blot (1:300)
Alternate names:
Rotamase A; Cyclosporin A-binding protein; Cyclophilin A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1025 375.00
Check your price
  • LAMP2 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA517868
  • Size 300ug
Applications
Flow Cytometry (1.0 ug/10^6 cells)
Alternate names:
Lysosomal membrane glycoprotein type B; CD107 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517868 375.00
Check your price
  • Erythrocytes Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA517824
  • Size 300ug
Applications
Flow Cytometry (0.5 ug/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517824 375.00
Check your price
  • GPR78 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132777
  • Size 25µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132777 375.00
Check your price
  • GPR133 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132849
  • Size 25µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
G-protein coupled receptor PGR25; G-protein coupled receptor 133
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132849 375.00
Check your price
  • GCNF Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132593
  • Size 25µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Retinoid receptor-related testis-specific receptor; Germ cell nuclear factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132593 375.00
Check your price
  • HSP70 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IP and GS
  • Host Mouse
  • Target Species Amphibian, Avian, Fruit fly, Fish, Flatworm, Human, Mouse, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # MA3007
  • Size 250µL
Applications
Gel Shift (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1:250-1:1000)
Alternate names:
Hsp68; Heat shock 70 kDa protein 3; Heat shock 70 kDa protein 2; Heat shock 70 kDa protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3007 375.00
Check your price
  • Caspase 6 Monoclonal Antibody for Western Blot, IHC (P) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191593
  • Size 250µL
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunoprecipitation (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
Apoptotic protease Mch-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191593 375.00
Check your price
  • MCP-1 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Mouse
  • Target Species Human, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA121383
  • Size 250µg
Applications
ELISA (Assay dependent), Neutralization (0.5 µg/ml), Western Blot (0.1-5 µg/ml)
Alternate names:
Small-inducible cytokine A2; Monocyte secretory protein JE; Monocyte chemotactic protein 1; Monocyte chemotactic and activating factor; Monocyte chemoattractant protein 1; HC11
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121383 375.00
Check your price
  • IL-10 Monoclonal Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Rat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # M011B
  • Size 250µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Cytokine synthesis inhibitory factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
M011B 375.00
Check your price
  • Haloperidol Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA191883
  • Size 250µg
Applications
ELISA (Assay dependent)
Alternate names:
Aloperidin, Bioperidolo, Brotopon, Dozic, Duraperidol, Einalon S, Eukystol, Haldol, Halosten, Keselan, Linton, Peluces, Serenace, Serenase, Sigaperido
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191883 375.00
Check your price
  • G-CSF Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121362
  • Size 250µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Pluripoietin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121362 375.00
Check your price