Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  50  60  70  80  90  100  110  120  130  138  139  140  141  142  143  144  145  146  147  148  149  150  151  152  153  154  155  156  157  158  159  160  161  162  163  164  165  166  167  170  180  190  200  210  220  230  240  250  300  400  500  600  700  800  900  1000  1100  1630 
4561 - 4590 of 48877 products displayed
  • Estrogen Receptor alpha Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA1309
  • Size 50µg
Applications
Immunocytochemistry (1:1000-1:4000), Immunofluorescence (1:1000-1:4000), Immunohistochemistry (Paraffin) (1-5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1309 375.00
Check your price
  • Eotaxin 2 Polyclonal Antibody for Western Blot, ELISA, Neu, FN and IA
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA125517
  • Size 50µg
Applications
ELISA (0.5 µg/ml), Functional Assay (7-8 ug/ml), Inhibition Assays (7-8 ug/ml), Neutralization (7-8 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
Small-inducible cytokine A24; Eotaxin-2; Eosinophil chemotactic protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125517 375.00
Check your price
  • CYR61 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA123143
  • Size 50µg
Applications
Western Blot (1-4 µg/ml)
Alternate names:
Insulin-like growth factor-binding protein 10; Cysteine-rich angiogenic inducer 61; CCN family member 1; 3CH61; Protein GIG1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123143 375.00
Check your price
  • CCR1 Polyclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120688
  • Size 50µg
Applications
ELISA (1:100,000), Immunocytochemistry (1:400), Immunofluorescence (1:400), Western Blot (1:1000)
Alternate names:
RANTES-R; Macrophage inflammatory protein 1-alpha receptor; LD78 receptor; HM145
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120688 375.00
Check your price
  • Aromatase Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Horse, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA121398
  • Size 50µg
Applications
Immunohistochemistry (Paraffin) (10-20 µg/ml), Western Blot (1-4 µg/ml)
Alternate names:
Estrogen synthase; Cytochrome P450 19A1; Cytochrome P-450AROM; CYPXIX; Cytochrome P450 19 type I; CYPXIXA1; Cytochrome P450 17-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121398 375.00
Check your price
  • Rat IgG2a/IgG1 Isotype Control, FITC, PE conjugate for Flow and Ctrl
  • Host Rat
  • Target Species Not Applicable
  • Conjugate FITC , PE
  • Catalog # PA533184
  • Size 50TESTS
Applications
Control (Assay-Dependent), Flow Cytometry (Neat)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533184 375.00
Check your price
  • Mouse IgG1/Rat IgG1 Isotype Control, FITC, PE conjugate for Flow and Ctrl
  • Host Mixed
  • Target Species Not Applicable
  • Conjugate FITC , PE
  • Catalog # PA533183
  • Size 50TESTS
Applications
Control (Assay-Dependent), Flow Cytometry (Neat)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533183 375.00
Check your price
  • XRCC1 Monoclonal Antibody for Western Blot, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA513412
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
X-ray repair cross-complementing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513412 375.00
Check your price
  • URO5 Monoclonal Antibody for IF, ICC and IHC (F)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA125301
  • Size 500µL
Applications
Immunocytochemistry (1:40), Immunofluorescence (1:40), Immunohistochemistry (Frozen) (1:40)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125301 375.00
Check your price
  • URO10 Monoclonal Antibody for IF, ICC and IHC (F)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA190564
  • Size 500µL
Applications
Immunocytochemistry (1:40), Immunofluorescence (1:40), Immunohistochemistry (Frozen) (1:40)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190564 375.00
Check your price
  • Rhodopsin Monoclonal Antibody, Biotin conjugate for Western Blot, IF and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA511738
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:25-1:50), Western Blot (1-2 µg/ml)
Alternate names:
Opsin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511738 375.00
Check your price
  • Progesterone Receptor Monoclonal Antibody for Western Blot, IF, IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Chicken, Horse, Human, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA512642
  • Size 500µL
Applications
Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin) (2-4 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512642 375.00
Check your price
  • PMP22 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191698
  • Size 500µL
Applications
Western Blot (Assay dependent)
Alternate names:
Growth arrest-specific protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191698 375.00
Check your price
  • Placental Alkaline Phosphatase Monoclonal Antibody, Biotin conjugate for IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA512691
  • Size 500µL
Applications
ELISA (Assay Dependent), Immunohistochemistry (1:200)
Alternate names:
Placental alkaline phosphatase 1; Alkaline phosphatase Regan isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512691 375.00
Check your price
  • p53 Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Bovine, Human, Non-human primate
  • Conjugate Biotin
  • Catalog # MA512554
  • Size 500µL
Applications
Flow Cytometry (0.5ug/test), Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Paraffin) (1:100-1:200 ), Immunoprecipitation (1:50-1:100 ), Western Blot (1:50-1:200)
Alternate names:
Tumor suppressor p53; Phosphoprotein p53; Antigen NY-CO-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512554 375.00
Check your price
  • Milk Fat Globule 1 Monoclonal Antibody, Biotin conjugate for Western Blot, IF, IHC (P) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA513553
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (0.5-1.0 µg/ml)
Alternate names:
SED1; Milk fat globule-EGF factor 8; MFGM; HMFG; Breast epithelial antigen BA46
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513553 375.00
Check your price
  • MBNL1 Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA116967
  • Size 500µL
Applications
Immunocytochemistry (1:50-1:250), Immunofluorescence (1:50-1:250), Immunohistochemistry (Paraffin) (1:1000), Western Blot (1:1000-1:5000)
Alternate names:
Triplet-expansion RNA-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116967 375.00
Check your price
  • MAP1B Monoclonal Antibody, Biotin conjugate for IF and IHC (P)
  • Host Mouse
  • Target Species Bovine, Human, Rat
  • Conjugate Biotin
  • Catalog # MA512813
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1-2 µg/ml)
Alternate names:
Neuraxin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512813 375.00
Check your price
  • Laminin beta-2/gamma-1 Monoclonal Antibody, Biotin conjugate for IF, IHC (F), IHC (P) and IP
  • Host Rat
  • Target Species Human, Mouse
  • Conjugate Biotin
  • Catalog # MA514646
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent), Immunoprecipitation (1:25)
Alternate names:
S-LAM beta, S-laminin subunit beta, laminin B1s chain, laminin S, laminin subunit beta-2, laminin-11 subunit beta, laminin-14 subunit beta, laminin-15 subunit beta, laminin-3 subunit beta, laminin-4 subunit beta, laminin-7 subunit beta, laminin-9 subunit beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514646 375.00
Check your price
  • Laminin beta-1 Monoclonal Antibody, Biotin conjugate for Western Blot, IF, IHC (F), IHC (P) and IP
  • Host Rat
  • Target Species Hamster, Human, Mouse, Pig
  • Conjugate Biotin
  • Catalog # MA514654
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (2 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Laminin-8 subunit beta; Laminin-6 subunit beta; Laminin-2 subunit beta; Laminin-12 subunit beta; Laminin-10 subunit beta; Laminin-1 subunit beta; Laminin B1 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514654 375.00
Check your price
  • GM-CSF Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132122
  • Size 500µL
Applications
ELISA (1:1000-1:5000), Western Blot (1:500-1:2000)
Alternate names:
Sargramostim; Molgramostin; Colony-stimulating factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132122 375.00
Check your price
  • Fibronectin Polyclonal Antibody for IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA123693
  • Size 500µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1:100)
Alternate names:
Cold-insoluble globulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123693 375.00
Check your price
  • Calcitonin Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516325
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Calcitonin gene-related peptide I; Alpha-type CGRP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516325 375.00
Check your price
  • Plasminogen Monoclonal Antibody for IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120080
  • Size 500µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
RP1-81D8.1, DKFZp779M0222
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120080 375.00
Check your price
  • Neutrophil elastase Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110606
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
PMN elastase; Medullasin; Human leukocyte elastase; Elastase-2; Bone marrow serine protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110606 375.00
Check your price
  • Neutrophil elastase Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110608
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
PMN elastase; Medullasin; Human leukocyte elastase; Elastase-2; Bone marrow serine protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110608 375.00
Check your price
  • Lidocaine Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA110751
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
Lignocaine
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110751 375.00
Check your price
  • IFN gamma Monoclonal Antibody for IF, ICC and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523718
  • Size 500µg
Applications
Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Neutralization (0.02-0.06 µg/ml)
Alternate names:
Immune interferon
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523718 375.00
Check your price
  • FABP4 Polyclonal Antibody for Western Blot and ELISA
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA121216
  • Size 500µg
Applications
ELISA (1:1000-1:5000), Western Blot (1:1000-1:5000)
Alternate names:
Fatty acid-binding protein 4; Adipocyte-type fatty acid-binding protein; Adipocyte lipid-binding protein; Protein 422; P2 adipocyte protein; P15; Myelin P2 protein homolog; 3T3-L1 lipid-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121216 375.00
Check your price
  • CXCL8 Monoclonal Antibody for Western Blot, IF, ICC, Flow, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523697
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Neutralization (Assay-Dependent), Western Blot (1 µg/ml)
Alternate names:
T-cell chemotactic factor; Protein 3-10C; Neutrophil-activating protein 1; Monocyte-derived neutrophil-activating peptide; Monocyte-derived neutrophil chemotactic factor; Granulocyte chemotactic protein 1; Emoctakin; Chemokine (C-X-C motif) ligand 8; C-X-C motif chemokine 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523697 375.00
Check your price