Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  40  50  60  70  80  90  100  110  200  300  400  500  600  700  800  900  1000  1605 
451 - 480 of 48136 products displayed
  • hCG Antibody (Monoclonal, SC1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIH9812
  • Size 1 mg
Applications
ELISA (2-10µg/ml), Immunohistochemistry (Paraffin) (1:20), Immunoprecipitation (5µg/ml), Radioimmune Assays (Assay Dependent), Western Blot (1:250-1:500)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9812 600.00
Check your price
  • Follicle Stimulating Hormone Antibody (Monoclonal, P2H3)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIF2710
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Follitropin beta chain; Follicle-stimulating hormone beta subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIF2710 600.00
Check your price
  • Ly-6B.2 Antibody, PE (Monoclonal, 7/4)
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA516542
  • Size 500 TESTS
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516542 600.00
Check your price
  • Trypsin Antibody (Monoclonal, 7C8)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB0210302
  • Size 200 µL
Applications
ELISA (1:8000), Western Blot (Assay dependent)
Alternate names:
Trypsin I; Serine protease 1; Cationic trypsinogen; Beta-trypsin
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB0210302 600.00
Check your price
  • Influenza A NP Antibody (Monoclonal, 5D8)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # HYB1560102
  • Size 200 µl
Applications
ELISA (Assay dependent), Immunocytochemistry (1:1000), Immunofluorescence (1:1,000), Western Blot (Assay dependent)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1560102 600.00
Check your price
  • Exendin 4 Antibody (Monoclonal, 20)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # ABS0122002
  • Size 200 µl
Applications
ELISA (1:32000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0122002 600.00
Check your price
  • Clostridium tetani Tetanus Toxin Antibody (Monoclonal, 10.D6)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # HYB2781002
  • Size 200 µl
Applications
ELISA (1:16000)
Alternate names:
C. tetani, pE88_60
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB2781002 600.00
Check your price
  • Parainfluenza Virus Type 1 Fusion Protein Antibody, FITC (Monoclonal, PAR1 (251/12A))
  • Host Mouse
  • Target Species Virus
  • Conjugate FITC
  • Catalog # MA516282
  • Size 2 mg
Applications
Immunofluorescence (Assay Dependent)
Alternate names:
hPIV-1, PIV-1, Parainfluenza Virus Type 1, hPIV-1 Fusion protein, PIV-1 Fusion protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516282 600.00
Check your price
  • Astrovirus Type 2 Antibody (Monoclonal, 8E7)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA516293
  • Size 2 mg
Applications
ELISA (Assay Dependent), Lateral Flow (Assay Dependent)
Alternate names:
Astro virus, Astroviridae
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516293 600.00
Check your price
  • IkB alpha Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA185928
  • Size 100 µL
Applications
ELISA (1:20,000-1:100,000), Immunohistochemistry (Frozen) (1/1000-1/5000), Immunoprecipitation (Assay Dependent), Western Blot (1:2000-1:10,000)
Alternate names:
Major histocompatibility complex enhancer-binding protein MAD3; I-kappa-B-alpha; RL/IF-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185928 600.00
Check your price
  • PR3 Antibody (Monoclonal, PR3-G2)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA140218
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent.), Immunohistochemistry (Frozen) (Assay Dependent.), Western Blot (Assay Dependent.)
Alternate names:
Wegener autoantigen; P29; Neutrophil proteinase 4; Leukocyte proteinase 3; C-ANCA antigen; AGP7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140218 600.00
Check your price
  • ASGR1 Antibody (Monoclonal, 8D7)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA140244
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent.), Immunofluorescence (Assay dependent), Immunohistochemistry (Frozen) (Assay Dependent.), Western Blot (Assay Dependent.)
Alternate names:
Hepatic lectin 1; Hepatic lectin H1; C-type lectin domain family 4 member H1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140244 600.00
Check your price
  • Yellow Fever Antibody (Monoclonal, 3576)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA516501
  • Size 100 ug
Applications
ELISA (1:20-1:200)
Alternate names:
YFV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516501 600.00
Check your price
  • RT1.B Antibody (Monoclonal, OX-6)
  • Host Mouse
  • Target Species Rat, Mouse
  • Conjugate Unconjugated
  • Catalog # MA180672
  • Size 1 mg
Applications
Flow Cytometry (1:50-1:100), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (1:50-1:100), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180672 600.00
Check your price
  • Fibronectin Antibody (Monoclonal, 3F12)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIF2601
  • Size 1 mg
Applications
ELISA (1 µg/ml), Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1:100-1:500)
Alternate names:
Cold-insoluble globulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIF2601 600.00
Check your price
  • IL-12 p70 Antibody (Monoclonal, CC301)
  • Host Mouse
  • Target Species Bovine, Human, Sheep
  • Conjugate Unconjugated
  • Catalog # MA516618
  • Size 500 ug
Applications
ELISA (8 ug/ml), Flow Cytometry (1:10), Functional Assay (1:100), Western Blot (1:50-1:100)
Alternate names:
NK cell stimulatory factor chain 1; IL-12 subunit p35; Cytotoxic lymphocyte maturation factor 35 kDa subunit; IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit; NK cell stimulatory factor chain 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516618 600.00
Check your price
  • Dectin 1 (soluble) Antibody (Monoclonal, 2A11)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA516478
  • Size 500 ug
Applications
Flow Cytometry (1:10 -1:50), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
BGR, Clecsf12, beta-GR,C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 12; C-type lectin domain family 7 member A; C-type lectin superfamily member 12; DC-associated C-type lectin 1; beta-glucan receptor; dectin-1; dendritic cell-associated C-type lectin 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516478 600.00
Check your price
  • Follicle Stimulating Hormone Antibody
  • Host Rabbit
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # PA185444
  • Size 50 µL
Applications
Immunohistochemistry (Frozen) (1/200-1/500), Western Blot (1/300-1/400)
Alternate names:
Follitropin beta chain; Follicle-stimulating hormone beta subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185444 600.00
Check your price
  • CD8 beta Antibody, PE (Monoclonal, CT-CD8b)
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA517850
  • Size 300 ug
Applications
Flow Cytometry (0.1 ug/10^6 cells)
Alternate names:
T-cell surface glycoprotein Lyt-3; T-cell membrane glycoprotein Ly-3; Lymphocyte antigen 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517850 600.00
Check your price
  • CD42a Antibody (Monoclonal, FMC-25)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516684
  • Size 250 ug
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Glycoprotein 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516684 600.00
Check your price
  • Trypsin Antibody (Monoclonal, 2C4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB0210802
  • Size 200 µl
Applications
ELISA (1:8000), Western Blot (Assay dependent)
Alternate names:
Trypsin I; Serine protease 1; Cationic trypsinogen; Beta-trypsin
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB0210802 600.00
Check your price
  • Mycobacterium tuberculosis ESAT-6 Antibody (Monoclonal, 11G4)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # HYB0760802
  • Size 200 µl
Applications
ELISA (1:3,200), Western Blot (Assay dependent)
Alternate names:
M. tuberculosis, myobacter
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB0760802 600.00
Check your price
  • Estradiol Antibody (Monoclonal, 7G4)
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # HYB0570202
  • Size 200 µl
Applications
ELISA (1:8000)
Alternate names:
17-beta-hydroxysteroid dehydrogenase, 17-beta-HSD, Estrogen 17-oxidoreductase
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB0570202 600.00
Check your price
  • Complement C5b Antibody (Monoclonal, aE11)
  • Host Mouse
  • Target Species Horse, Human, Non-human primate, Pig
  • Conjugate Unconjugated
  • Catalog # DIA0110102
  • Size 200 µl
Applications
ELISA (1/20,000), Immunohistochemistry (Assay Dependent)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
DIA0110102 600.00
Check your price
  • Aprotinin Antibody (Monoclonal, 18)
  • Host Mouse
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # ABS0371802
  • Size 200 µL
Applications
ELISA (0.6-20 ug/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0371802 600.00
Check your price
  • TSH Receptor Antibody (Monoclonal, 4C1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516519
  • Size 200 ug
Applications
Flow Cytometry (1:100 -1:1000), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1:1000-1:2000), Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
Thyroid-stimulating hormone receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516519 600.00
Check your price
  • Parainfluenza Virus Type 3 HA Antibody, FITC (Monoclonal, PAR3 (341/7H))
  • Host Mouse
  • Target Species Virus
  • Conjugate FITC
  • Catalog # MA516280
  • Size 2 mg
Applications
Immunofluorescence (Assay Dependent)
Alternate names:
hPIV-3, PIV-3, Parainfluenza Virus Type 3, PIV-3 hemagglutinin-Neuraminidase, HPIV3gp7, PIV-3 hemagglutinin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516280 600.00
Check your price
  • HSV Type 2 Antibody (Monoclonal, AP1)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA516286
  • Size 2 mg
Applications
Immunofluorescence (Assay Dependent)
Alternate names:
HSV2, HSV-2, Human herpes virus 2, HHV-2, Herpes Simplex II, HSV Type 2, Herpes Simplex 2, HSV Type II
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516286 600.00
Check your price
  • Herpes Simplex Virus Type 1/2 Antibody (Monoclonal, AP7)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA516294
  • Size 2 mg
Applications
ELISA (Assay Dependent)
Alternate names:
HSV1, Herpes Simplex I, HSV Type 1, Herpes Simplex 1, HHV, HSV, Herpes Simplex Type 1, HSV2, HSV-2, Human herpes virus 2, HHV-2, Herpes Simplex II, HSV Type 2, Herpes Simplex 2, HSV Type II, HSV 1/2, HSV I/II
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516294 600.00
Check your price
  • LSD Antibody
  • Host Sheep
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # PA175244
  • Size 100 µL
Applications
ELISA (Assay Dependent)
Alternate names:
Lysergic Acid Diethylamide
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175244 600.00
Check your price