Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  60  70  80  90  100  110  120  130  140  146  147  148  149  150  151  152  153  154  155  156  157  158  159  160  161  162  163  164  165  166  167  168  169  170  171  172  173  174  175  180  190  200  210  220  230  240  250  260  300  400  500  600  700  800  900  1000  1100  1602 
4801 - 4830 of 48059 products displayed
  • Imipramine Antibody (Monoclonal, IM11-214.1)
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA110722
  • Size 500 µg
Applications
ELISA (Assay dependent)
Alternate names:
Antideprin®, Deprenil, Deprimin, Deprinol, Depsonil, Dynaprin, Eupramin, Imipramil, Irmin, Janimine®, Melipramin, Surplix, Tofranil
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110722 375.00
Check your price
  • IL-8 (CXCL8) Antibody (Monoclonal, 6217)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523697
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Neutralization (Assay-Dependent), Western Blot (1 µg/ml)
Alternate names:
T-cell chemotactic factor; Protein 3-10C; Neutrophil-activating protein 1; Monocyte-derived neutrophil-activating peptide; Monocyte-derived neutrophil chemotactic factor; Granulocyte chemotactic protein 1; Emoctakin; Chemokine (C-X-C motif) ligand 8; C-X-C motif chemokine 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523697 375.00
Check your price
  • IL-7 Antibody (Monoclonal, 7417)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523700
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.4-0.8 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Interleukin-7, IL7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523700 375.00
Check your price
  • IL-6 Antibody (Monoclonal, MP520F3)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523800
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.005-0.025 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Interleukin HP-1; B-cell hybridoma growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523800 375.00
Check your price
  • IL-6 Antibody (Monoclonal, 6708)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523698
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.05-0.15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Interferon beta-2; Hybridoma growth factor; CTL differentiation factor; B-cell stimulatory factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523698 375.00
Check your price
  • IL-6 Antibody (Monoclonal, 5IL6)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # M620
  • Size 500 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Interferon beta-2; Hybridoma growth factor; CTL differentiation factor; B-cell stimulatory factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
M620 375.00
Check your price
  • IL-22 Antibody (Monoclonal, 142906)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523775
  • Size 500 µg
Applications
ELISA (0.5-2 µg/ml (Detection), 2-8 µg/ml (Capture))
Alternate names:
IL-10-related T-cell-derived-inducible factor; Cytokine Zcyto18
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523775 375.00
Check your price
  • IL-2 Antibody (Monoclonal, 5355)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523696
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml)
Alternate names:
T-cell growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523696 375.00
Check your price
  • IL-17A Antibody (Monoclonal, 50101)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523747
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Western Blot (1 µg/ml)
Alternate names:
Cytotoxic T-lymphocyte-associated antigen 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523747 375.00
Check your price
  • IL-12 p70 Antibody (Monoclonal, 20C2)
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # M122
  • Size 500 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
NK cell stimulatory factor chain 1; IL-12 subunit p35; Cytotoxic lymphocyte maturation factor 35 kDa subunit; NK cell stimulatory factor chain 2; IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
M122 375.00
Check your price
  • IL-11 Antibody (Monoclonal, 22616)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523711
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Western Blot (1 µg/ml)
Alternate names:
Adipogenesis inhibitory factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523711 375.00
Check your price
  • IL-10 Antibody (Monoclonal, DNAX JES52A5)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523796
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.005-0.015 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Cytokine synthesis inhibitory factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523796 375.00
Check your price
  • IL-1 beta Antibody (Monoclonal, 30311)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523721
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (1-5 µg/ml)
Alternate names:
IL-1 beta, catabolin, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta, IL1B, IL1 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523721 375.00
Check your price
  • IGFBP1 Antibody (Monoclonal, 33627)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523727
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (10-40 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Placental protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523727 375.00
Check your price
  • Hemoglobin Antibody (Monoclonal, HB11-201.11)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110806
  • Size 500 µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Hemoglobin alpha chain; Alpha-globin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110806 375.00
Check your price
  • hCG beta Antibody (Monoclonal, INN-hCG-68)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA182291
  • Size 500 µg
Applications
ELISA (1/1000-1/80000), Radioimmune Assays (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182291 375.00
Check your price
  • Gram Negative Endotoxin Antibody (Monoclonal, GNE13-337.5)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA110684
  • Size 500 µg
Applications
ELISA (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110684 375.00
Check your price
  • Gram Negative Endotoxin Antibody (Monoclonal, GNE11-270.3.1)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA110688
  • Size 500 µg
Applications
ELISA (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110688 375.00
Check your price
  • Gram Negative Endotoxin Antibody (Monoclonal, GNE11-265.5)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA110687
  • Size 500 µg
Applications
ELISA (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110687 375.00
Check your price
  • Goat anti-Armenian Hamster IgG, HRP
  • Host Goat
  • Target Species Hamster
  • Conjugate HRP
  • Catalog # PA132045
  • Size 500 µg
Applications
Dot blot (Assay dependent), ELISA (1:4000-1:20,000), Immunomicroscopy (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132045 373.00
Check your price
  • GnRH1 Antibody (Monoclonal, LHRH13-426.12)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110817
  • Size 500 µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Assay dependent), Radioimmune Assays (Assay dependent)
Alternate names:
Progonadoliberin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110817 375.00
Check your price
  • FABP4 Antibody
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA121216
  • Size 500 µg
Applications
ELISA (1:1000-1:5000), Western Blot (1:1000-1:5000)
Alternate names:
Fatty acid-binding protein 4; Adipocyte-type fatty acid-binding protein; Adipocyte lipid-binding protein; Protein 422; P2 adipocyte protein; P15; Myelin P2 protein homolog; 3T3-L1 lipid-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121216 375.00
Check your price
  • FABP3 Antibody (Monoclonal, 10E1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA122721
  • Size 500 µg
Applications
ELISA (Assay dependent)
Alternate names:
Muscle fatty acid-binding protein; Mammary-derived growth inhibitor; Heart-type fatty acid-binding protein; Fatty acid-binding protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122721 375.00
Check your price
  • EGF Antibody (Monoclonal, 10827)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523704
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Immunohistochemistry (8-25 µg/ml)
Alternate names:
Epidermal Growth Factor, URG, Urogastrone, HOMG4, URG, beta-urogastrone, pro-epidermal growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523704 375.00
Check your price
  • Digoxin Antibody (Monoclonal, DG D8-9)
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA110809
  • Size 500 µg
Applications
ELISA (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110809 375.00
Check your price
  • Digoxin Antibody (Monoclonal, DG D46)
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA110811
  • Size 500 µg
Applications
ELISA (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110811 375.00
Check your price
  • CXCL2 Antibody (Monoclonal, 40605)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523737
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (1-4 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Macrophage inflammatory protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523737 375.00
Check your price
  • CXCL10 Antibody (Monoclonal, 33036)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523726
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Neutralization (0.5-2.0 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Small-inducible cytokine B10; 10 kDa interferon gamma-induced protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523726 375.00
Check your price
  • CXCL1 Antibody (Monoclonal, 20326)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523707
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Neutralization (3-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Neutrophil-activating protein 3; Melanoma growth stimulatory activity; GRO-alpha(1-73); C-X-C motif chemokine 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523707 375.00
Check your price
  • Chymotrypsin Antibody (Monoclonal, CHY 10-104.5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110623
  • Size 500 µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Caldecrin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110623 375.00
Check your price