Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  60  70  80  90  100  110  120  130  140  151  152  153  154  155  156  157  158  159  160  161  162  163  164  165  166  167  168  169  170  171  172  173  174  175  176  177  178  179  180  190  200  210  220  230  240  250  260  300  400  500  600  700  800  900  1000  1100  1630 
4951 - 4980 of 48882 products displayed
  • TNF alpha Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # M303
  • Size 500µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Tumor necrosis factor ligand superfamily member 2; TNF-alpha; Cachectin
Catalog # Size Price (AUD) Your Price (AUD) Qty
M303 375.00
Check your price
  • TNF alpha Monoclonal Antibody for Western Blot, IF, ICC, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523720
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Neutralization (0.04-0.08 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Tumor necrosis factor ligand superfamily member 2; TNF-alpha; Cachectin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523720 375.00
Check your price
  • TIMP1 Monoclonal Antibody for Western Blot, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523756
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Tissue inhibitor of metalloproteinases 1; Fibroblast collagenase inhibitor; Erythroid-potentiating activity
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523756 375.00
Check your price
  • Theophylline Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA110735
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
Dimethylxanthine
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110735 375.00
Check your price
  • TGF beta-1,2,3 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), ELISA, Neu and IA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523795
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Immunohistochemistry (Paraffin) (8-25 µg/ml), Inhibition Assays (Assay-Dependent), Neutralization (0.25-1.25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
TGFB, TGFb-1, DPD1, Transforming Growth Factor, Transforming Growth Factor Beta-1, TGF-beta-1, CED
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523795 375.00
Check your price
  • TGF beta-1 Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA and Neu
  • Host Mouse
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523702
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (Assay-Dependent), Neutralization (0.3-1.0 µg/ml), Western Blot (1 µg/ml)
Alternate names:
TGFB, TGFb-1, DPD1, Transforming Growth Factor, Transforming Growth Factor Beta-1, TGF-beta-1, CED
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523702 375.00
Check your price
  • TGF alpha Polyclonal Antibody for ELISA
  • Host Sheep
  • Target Species Bovine, Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA185931
  • Size 500µg
Applications
ELISA (Assay Dependent)
Alternate names:
TFGA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185931 375.00
Check your price
  • S100B Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA123052
  • Size 500µg
Applications
ELISA (Assay Dependent), Western Blot (Assay dependent)
Alternate names:
S100 calcium-binding protein B; S-100 protein subunit beta; S-100 protein beta chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123052 375.00
Check your price
  • Quinidine Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA191521
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191521 375.00
Check your price
  • Plasminogen Monoclonal Antibody for IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120083
  • Size 500µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
RP1-81D8.1, DKFZp779M0222
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120083 375.00
Check your price
  • NTF3 Polyclonal Antibody for Western Blot, IHC, ELISA and IA
  • Host Rabbit
  • Target Species Human, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA118391
  • Size 500µg
Applications
ELISA (1-10 µg/ml), Immunohistochemistry (1-10 µg/ml), Inhibition Assays (1-10 µg/ml), Western Blot (1-10 µg/ml)
Alternate names:
Neurotrophic factor; Nerve growth factor 2; HDNF
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118391 375.00
Check your price
  • NTF3 Polyclonal Antibody for Western Blot, IHC, ELISA and IA
  • Host Sheep
  • Target Species Human, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA118389
  • Size 500µg
Applications
ELISA (1-10 µg/ml), Immunohistochemistry (1-10 µg/ml), Inhibition Assays (1-10 µg/ml), Western Blot (1-10 µg/ml)
Alternate names:
Neurotrophic factor; Nerve growth factor 2; HDNF
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118389 375.00
Check your price
  • NGF Polyclonal Antibody for Western Blot, IHC, ELISA and IA
  • Host Sheep
  • Target Species Avian, Chicken, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA118381
  • Size 500µg
Applications
ELISA (1-3 µg/ml), Immunohistochemistry (1-3 µg/ml), Inhibition Assays (1-3 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
beta Nerve Growth Factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118381 375.00
Check your price
  • Neutrophil elastase Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110607
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
PMN elastase; Medullasin; Human leukocyte elastase; Elastase-2; Bone marrow serine protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110607 375.00
Check your price
  • Neutrophil elastase Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110608
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
PMN elastase; Medullasin; Human leukocyte elastase; Elastase-2; Bone marrow serine protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110608 375.00
Check your price
  • Imipramine Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA110722
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
Antideprin®, Deprenil, Deprimin, Deprinol, Depsonil, Dynaprin, Eupramin, Imipramil, Irmin, Janimine®, Melipramin, Surplix, Tofranil
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110722 375.00
Check your price
  • IL-6 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523800
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.005-0.025 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Interleukin HP-1; B-cell hybridoma growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523800 375.00
Check your price
  • IL-6 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523698
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.05-0.15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Interferon beta-2; Hybridoma growth factor; CTL differentiation factor; B-cell stimulatory factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523698 375.00
Check your price
  • IL-6 Monoclonal Antibody for Western Blot, Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # M620
  • Size 500µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Interferon beta-2; Hybridoma growth factor; CTL differentiation factor; B-cell stimulatory factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
M620 375.00
Check your price
  • IL-22 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523775
  • Size 500µg
Applications
ELISA (0.5-2 µg/ml (Detection), 2-8 µg/ml (Capture))
Alternate names:
IL-10-related T-cell-derived-inducible factor; Cytokine Zcyto18
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523775 375.00
Check your price
  • IL-17A Monoclonal Antibody for Western Blot and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523747
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Western Blot (1 µg/ml)
Alternate names:
Cytotoxic T-lymphocyte-associated antigen 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523747 375.00
Check your price
  • IL-12 p70 Monoclonal Antibody for Western Blot and ELISA
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # M122
  • Size 500µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
NK cell stimulatory factor chain 1; IL-12 subunit p35; Cytotoxic lymphocyte maturation factor 35 kDa subunit; NK cell stimulatory factor chain 2; IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
M122 375.00
Check your price
  • IL-11 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523711
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Western Blot (1 µg/ml)
Alternate names:
Adipogenesis inhibitory factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523711 375.00
Check your price
  • IL-1 beta Monoclonal Antibody for ELISA and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523721
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (1-5 µg/ml)
Alternate names:
IL-1 beta, catabolin, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta, IL1B, IL1 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523721 375.00
Check your price
  • IL-1 beta Monoclonal Antibody for Western Blot, IF, ICC, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523691
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Neutralization (0.05-0.2 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523691 375.00
Check your price
  • IGFBP1 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523727
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (10-40 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Placental protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523727 375.00
Check your price
  • IFN gamma Monoclonal Antibody for IF, ICC and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523718
  • Size 500µg
Applications
Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Neutralization (0.02-0.06 µg/ml)
Alternate names:
Immune interferon
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523718 375.00
Check your price
  • Hemoglobin Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110806
  • Size 500µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Hemoglobin alpha chain; Alpha-globin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110806 375.00
Check your price
  • hCG beta Monoclonal Antibody for Western Blot, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA182291
  • Size 500µg
Applications
ELISA (1/1000-1/80000), Radioimmune Assays (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182291 375.00
Check your price
  • Gram Negative Endotoxin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA110684
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110684 375.00
Check your price