Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  70  80  90  100  110  120  130  140  150  155  156  157  158  159  160  161  162  163  164  165  166  167  168  169  170  171  172  173  174  175  176  177  178  179  180  181  182  183  184  190  200  210  220  230  240  250  260  300  400  500  600  700  800  900  1000  1100  1630 
5071 - 5100 of 48877 products displayed
  • HDAC2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532444
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (1:25)
Alternate names:
HD2, RPD3, YAF1, YY1-associated factor 1, transcriptional regulator homolog RPD3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532444 375.00
Check your price
  • gamma Catenin Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA120930
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Desmoplakin-3; Desmoplakin III; Catenin gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120930 375.00
Check your price
  • ErbB3 Polyclonal Antibody for IHC (P) and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130497
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Immunoprecipitation (Assay dependent)
Alternate names:
Tyrosine kinase-type cell surface receptor HER3; Proto-oncogene-like protein c-ErbB-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130497 375.00
Check your price
  • EpCAM Monoclonal Antibody, Biotin conjugate for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA512150
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml)
Alternate names:
Tumor-associated calcium signal transducer 1; Major gastrointestinal tumor-associated protein GA733-2; KSA; KS 1/4 antigen; Epithelial glycoprotein 314; Epithelial glycoprotein; Epithelial cell surface antigen; Cell surface glycoprotein Trop-1; Adenocarcinoma-associated antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512150 375.00
Check your price
  • EGFR Monoclonal Antibody for IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516359
  • Size 500µL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516359 375.00
Check your price
  • EGFR Monoclonal Antibody, Biotin conjugate for IF, IHC (F), Flow and IP
  • Host Mouse
  • Target Species Horse, Human
  • Conjugate Biotin
  • Catalog # MA513045
  • Size 500µL
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (2 µg/ml)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513045 375.00
Check your price
  • EGFR Monoclonal Antibody, Biotin conjugate for Flow and IP
  • Host Mouse
  • Target Species Dog, Human
  • Conjugate Biotin
  • Catalog # MA512872
  • Size 500µL
Applications
Flow Cytometry (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512872 375.00
Check your price
  • EGFR Monoclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513343
  • Size 500µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513343 375.00
Check your price
  • DP2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA129135
  • Size 500µL
Applications
Western Blot (1:1000)
Alternate names:
E2F dimerization partner 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129135 375.00
Check your price
  • DP1 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA129134
  • Size 500µL
Applications
Immunoprecipitation (1:2000), Western Blot (1:1000)
Alternate names:
E2F dimerization partner 1; DRTF1-polypeptide 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129134 375.00
Check your price
  • Cyclin D1 Monoclonal Antibody, Biotin conjugate for Western Blot, IF, IHC, Flow and IP
  • Host Mouse
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Biotin
  • Catalog # MA512699
  • Size 500µL
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
PRAD1 oncogene; BCL-1 oncogene; B-cell lymphoma 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512699 375.00
Check your price
  • Cyclin B1 Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA514324
  • Size 500µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (2-4 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1:100-1:1000)
Alternate names:
G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1,OTTHUMP00000223023
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514324 375.00
Check your price
  • Cyclin A2 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA121774
  • Size 500µL
Applications
Immunoprecipitation (10 µg/mg of lysate), Western Blot (5 µg/ml)
Alternate names:
CCN1, CCNA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121774 375.00
Check your price
  • COX2 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA514568
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (1-3 µg/mL)
Alternate names:
Prostaglandin-endoperoxide synthase 2; Prostaglandin H2 synthase 2; PHS II; Cyclooxygenase-2; TIS10 protein; PES-2; Macrophage activation-associated marker protein P71/73; Gripghs; Glucocorticoid-regulated inflammatory cyclooxygenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514568 375.00
Check your price
  • Connexin 43 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA120953
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (2-4 µg/ml)
Alternate names:
Gap junction 43 kDa heart protein; Connexin-43
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120953 375.00
Check your price
  • Claudin 3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA516867
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1-2 µg/mL)
Alternate names:
Rat ventral prostate.1 protein; Rat ventral prostate.1 protein homolog; Clostridium perfringens enterotoxin receptor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516867 375.00
Check your price
  • Cdc37 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA129131
  • Size 500µL
Applications
Immunoprecipitation (1:2000), Western Blot (1:1000)
Alternate names:
p50Cdc37; Hsp90 chaperone protein kinase-targeting subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129131 375.00
Check your price
  • CD90 Monoclonal Antibody for IF, ICC, IHC (F) and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA127016
  • Size 500µL
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (1:5000)
Alternate names:
Thy-1 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA127016 375.00
Check your price
  • CD8 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532334
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
T-lymphocyte differentiation antigen T8/Leu-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532334 375.00
Check your price
  • CD79a Monoclonal Antibody for IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516344
  • Size 500µL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (1:400)
Alternate names:
Surface IgM-associated protein; Membrane-bound immunoglobulin-associated protein; MB-1 membrane glycoprotein; Ig-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516344 375.00
Check your price
  • CD79a Monoclonal Antibody, Biotin conjugate for Western Blot, IF and IHC (P)
  • Host Mouse
  • Target Species Bovine, Chicken, Horse, Guinea pig, Human, Mouse, Non-human primate, Pig, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # MA513209
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (0.5-1 ug/ml), Western Blot (0.5-1.0 µg/ml)
Alternate names:
Surface IgM-associated protein; Membrane-bound immunoglobulin-associated protein; MB-1 membrane glycoprotein; Ig-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513209 375.00
Check your price
  • CD56 Monoclonal Antibody for IHC (F) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA190570
  • Size 500µL
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (1:25-1:50)
Alternate names:
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190570 375.00
Check your price
  • CD5 Monoclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514560
  • Size 500µL
Applications
Flow Cytometry (1:2), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (2-4 µg/ml)
Alternate names:
Lymphocyte antigen T1/Leu-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514560 375.00
Check your price
  • CD5 Monoclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513308
  • Size 500µL
Applications
Flow Cytometry (1:2), Immunohistochemistry (Paraffin) (1:20-1:40), Western Blot (1:10-1:200)
Alternate names:
Lymphocyte antigen T1/Leu-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513308 375.00
Check your price
  • CD205 Monoclonal Antibody, Biotin conjugate for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Biotin
  • Catalog # MA513362
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (2-4 µg/ml), Western Blot (Assay Dependent)
Alternate names:
CA-IX, P54/58N, RCC-associated antigen G250, RCC-associated protein G250, carbonate dehydratase IX, carbonic anhydrase 9, carbonic dehydratase, membrane antigen MN, pMW1, renal cell carcinoma-associated antigen G250
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513362 375.00
Check your price
  • CD2 Monoclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511373
  • Size 500µL
Applications
Flow Cytometry (2 ug/test), Immunohistochemistry (Paraffin) (1:30-1:60), Western Blot (1:100-1:500)
Alternate names:
T-cell surface antigen T11/Leu-5; Rosette receptor; LFA-3 receptor; LFA-2; Erythrocyte receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511373 375.00
Check your price
  • CD1a Monoclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA512526
  • Size 500µL
Applications
Flow Cytometry (2 ug/test), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:100-1:1000)
Alternate names:
T-cell surface antigen T6/Leu-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512526 375.00
Check your price
  • CD1a Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514096
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:15)
Alternate names:
T-cell surface antigen T6/Leu-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514096 375.00
Check your price
  • CD137 Monoclonal Antibody, Biotin conjugate for IF and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA513736
  • Size 500µL
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent)
Alternate names:
T-cell antigen ILA; T-cell antigen 4-1BB homolog; CDw137; 4-1BB ligand receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513736 375.00
Check your price
  • CD10 Monoclonal Antibody for Western Blot, IF, IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA514050
  • Size 500µL
Applications
Flow Cytometry (1:2), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (1:30-1:60), Western Blot (1:20-1:200)
Alternate names:
Skin fibroblast elastase; Neutral endopeptidase 24.11; Enkephalinase; Atriopeptidase; Common acute lymphocytic leukemia antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514050 375.00
Check your price