Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  80  90  100  110  120  130  140  150  160  168  169  170  171  172  173  174  175  176  177  178  179  180  181  182  183  184  185  186  187  188  189  190  191  192  193  194  195  196  197  200  210  220  230  240  250  260  270  280  300  400  500  600  700  800  900  1000  1100  1622 
5461 - 5490 of 48655 products displayed
  • SLC7A11 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA116893
  • Size 100 µL
Applications
Flow Cytometry (2-3 µg/ml), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (1:1000)
Alternate names:
xCT; Solute carrier family 7 member 11; Calcium channel blocker resistance protein CCBR1; Amino acid transport system xc-
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116893 375.00
Check your price
  • SIRT2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA3200
  • Size 100 µL
Applications
Immunocytochemistry (1:400), Immunofluorescence (1:400), Western Blot (1:1,000)
Alternate names:
SIR2-like protein 2; Regulatory protein SIR2 homolog 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3200 375.00
Check your price
  • Sir2.1 Polyclonal Antibody for Western Blot, IHC, IHC (P) and IP
  • Host Rabbit
  • Target Species C. elegans
  • Conjugate Unconjugated
  • Catalog # PA116933
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:200-1:500), Immunoprecipitation (1:10-1:500), Western Blot (1:3000)
Alternate names:
Regulatory protein SIR2 homolog 1; Protein sir-2.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116933 375.00
Check your price
  • SERPINB12 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA123493
  • Size 100 µL
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (1:1000)
Alternate names:
MGC119247, MGC119248, YUKOPIN
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123493 375.00
Check your price
  • SERCA2 ATPase Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Bovine, Dog, Human, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3910
  • Size 100 µL
Applications
Flow Cytometry (1µg per 10^6 cells), Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (1:500), Immunohistochemistry (Paraffin) (1 µg/ml), Western Blot (1:500-1:5000)
Alternate names:
Endoplasmic reticulum class 1/2 Ca(2+) ATPase; Calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletal muscle isoform; Calcium pump 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3910 375.00
Check your price
  • SERCA1 ATPase Monoclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Mouse
  • Target Species Dog, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3911
  • Size 100 µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (Frozen) (1:20-200), Western Blot (1:500-1:5000)
Alternate names:
Endoplasmic reticulum class 1/2 Ca(2+) ATPase; Calcium-transporting ATPase sarcoplasmic reticulum type, fast twitch skeletal muscle isoform; Calcium pump 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3911 375.00
Check your price
  • SEC61A1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA3014
  • Size 100 µL
Applications
Western Blot (1:20,000)
Alternate names:
Sec61 Complex Alpha Subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3014 375.00
Check your price
  • SCP3 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Chicken, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA116766
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Frozen) (1:10-1:500), Immunohistochemistry (Paraffin) (1:400), Western Blot (0.5 µg/ml)
Alternate names:
COR1, RNASCP3, Sycp3, Synaptonemal complex protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116766 375.00
Check your price
  • SAP97 Polyclonal Antibody for Western Blot, IF, ICC, IHC (F) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1741
  • Size 100 µL
Applications
Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Frozen) (1:1,000), Immunoprecipitation (Assay dependent), Western Blot (1:500-1:5000)
Alternate names:
Synapse-associated protein 97; Embryo-dlg/synapse-associated protein 97; hDlg
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1741 375.00
Check your price
  • S100B Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and ELISA
  • Host Mouse
  • Target Species Bovine, Dog, Cat, Goat, Human, Sheep, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA125005
  • Size 100 µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (1:100), Western Blot (Assay Dependent)
Alternate names:
S100 calcium-binding protein B; S-100 protein subunit beta; S-100 protein beta chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125005 375.00
Check your price
  • S100A2 Monoclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Bovine, Dog, Cat, Goat, Human, Pig, Reptile, Rat
  • Conjugate Unconjugated
  • Catalog # MA125006
  • Size 100 µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
S100 calcium-binding protein A2; Protein S-100L; CAN19
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125006 375.00
Check your price
  • S-arrestin Monoclonal Antibody, HRP conjugate for Western Blot, IHC and IHC (F)
  • Host Mouse
  • Target Species Bovine, Human, Pig
  • Conjugate HRP
  • Catalog # MA516030
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Western Blot (1:500-1:2000)
Alternate names:
Rod photoreceptor arrestin; Retinal S-antigen; 48 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516030 375.00
Check your price
  • S-arrestin Monoclonal Antibody, DyLight® 650 conjugate for Western Blot, IF, ICC, IHC and IHC (F)
  • Host Mouse
  • Target Species Bovine, Human, Pig
  • Conjugate DyLight® 650
  • Catalog # MA516028
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Western Blot (1:500-1:2000)
Alternate names:
Rod photoreceptor arrestin; Retinal S-antigen; 48 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516028 375.00
Check your price
  • S-arrestin Monoclonal Antibody, DyLight® 488 conjugate for Western Blot, IF, ICC, IHC and IHC (F)
  • Host Mouse
  • Target Species Bovine, Human, Pig
  • Conjugate DyLight® 488
  • Catalog # MA516029
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Western Blot (1:500-1:2000)
Alternate names:
Rod photoreceptor arrestin; Retinal S-antigen; 48 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516029 375.00
Check your price
  • S-arrestin Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC and IHC (F)
  • Host Mouse
  • Target Species Bovine, Human, Pig
  • Conjugate Biotin
  • Catalog # MA516027
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Western Blot (1:500-1:2000)
Alternate names:
Rod photoreceptor arrestin; Retinal S-antigen; 48 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516027 375.00
Check your price
  • RyR1 Monoclonal Antibody for Western Blot, IF, IHC (F) and IP
  • Host Mouse
  • Target Species Amphibian, Bovine, Dog, Chicken, Fish, Human, Mustelid, Mouse, Non-human primate, Sheep, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3925
  • Size 100 µL
Applications
Immunofluorescence (Assay dependent), Immunohistochemistry (Frozen) (1:1,000), Immunoprecipitation (Assay dependent), Western Blot (1:5,000)
Alternate names:
Type 1 ryanodine receptor; Skeletal muscle-type ryanodine receptor; Skeletal muscle ryanodine receptor; Skeletal muscle calcium release channel
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3925 375.00
Check your price
  • RUNX1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSR00271W
  • Size 100 µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
SL3/AKV core-binding factor alpha B subunit; SL3-3 enhancer factor 1 alpha B subunit; Polyomavirus enhancer-binding protein 2 alpha B subunit; Oncogene AML-1; Core-binding factor subunit alpha-2; Acute myeloid leukemia 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00271W 375.00
Check your price
  • RTN1 Monoclonal Antibody for Western Blot, ICC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA106810
  • Size 100 µL
Applications
Flow Cytometry (1:50-1:100), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Frozen) (1:50-1:100), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:50-1:500)
Alternate names:
Neuroendocrine-specific protein; S-rex
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106810 375.00
Check your price
  • rRNA Monoclonal Antibody, HRP conjugate for IP
  • Host Mouse
  • Target Species All
  • Conjugate HRP
  • Catalog # MA516063
  • Size 100 µL
Applications
Immunocytochemistry (1:2500), Immunofluorescence (1:2500), Immunoprecipitation (10 µl/lysate)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516063 375.00
Check your price
  • rRNA Monoclonal Antibody, DyLight® 488 conjugate for IF, ICC and IP
  • Host Mouse
  • Target Species All
  • Conjugate DyLight® 488
  • Catalog # MA516062
  • Size 100 µL
Applications
Immunocytochemistry (1:2500), Immunofluorescence (1:2500), Immunoprecipitation (10 µl/lysate)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516062 375.00
Check your price
  • rRNA Monoclonal Antibody, Biotin conjugate for IF, ICC and IP
  • Host Mouse
  • Target Species All
  • Conjugate Biotin
  • Catalog # MA516060
  • Size 100 µL
Applications
Immunocytochemistry (1:2500), Immunofluorescence (1:2500), Immunoprecipitation (10 µl/lysate)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516060 375.00
Check your price
  • rRNA Monoclonal Antibody for IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species All
  • Conjugate Unconjugated
  • Catalog # MA116628
  • Size 100 µL
Applications
Immunocytochemistry (1:2500), Immunofluorescence (1:2500), Immunohistochemistry (Paraffin) (Assay-Dependent), Immunoprecipitation (10 µl/lysate)
Alternate names:
ribosomal RNA 5.8s
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116628 375.00
Check your price
  • Recoverin Monoclonal Antibody, HRP conjugate for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate HRP
  • Catalog # MA516110
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
Cancer-associated retinopathy protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516110 375.00
Check your price
  • Recoverin Monoclonal Antibody, DyLight® 650 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human
  • Conjugate DyLight® 650
  • Catalog # MA516108
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
Cancer-associated retinopathy protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516108 375.00
Check your price
  • Recoverin Monoclonal Antibody, DyLight® 550 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human
  • Conjugate DyLight® 550
  • Catalog # MA516111
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
Cancer-associated retinopathy protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516111 375.00
Check your price
  • Recoverin Monoclonal Antibody, Biotin conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA516107
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
Cancer-associated retinopathy protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516107 375.00
Check your price
  • RAD21 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA116585
  • Size 100 µL
Applications
Western Blot (1:500)
Alternate names:
SCC1 homolog; Pokeweed agglutinin-binding protein 29; Nuclear matrix protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116585 375.00
Check your price
  • RAB9A Monoclonal Antibody for Western Blot, IF, ICC, IHC, IP and IM
  • Host Mouse
  • Target Species Bovine, Dog, Hamster, Human, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA3067
  • Size 100 µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (1:20), Immunomicroscopy (Assay Dependent), Immunoprecipitation (Assay dependent), Western Blot (1:1000-1:5000)
Alternate names:
Ras-related protein Rab-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3067 375.00
Check your price
  • PYY Polyclonal Antibody, Biotin conjugate for ELISA
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # PA19589
  • Size 100 µL
Applications
ELISA (1:2000-1:10000)
Alternate names:
Peptide tyrosine tyrosine; PYY-I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19589 375.00
Check your price
  • PYY Polyclonal Antibody, Biotin conjugate for ELISA
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # PA19590
  • Size 100 µL
Applications
ELISA (1:2000-1:10000)
Alternate names:
Peptide tyrosine tyrosine; PYY-I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19590 375.00
Check your price