Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  31  32  33  40  50  60  70  80  90  100  110  200  300  400  500  600  700  800  900  1000  1630 
541 - 570 of 48882 products displayed
  • Collagen II Monoclonal Antibody for Western Blot, IF, IHC (P) and Flow
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA137493
  • Size 200µg
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1-2 µg/ml for 30 min @ RT), Western Blot (1-2 µg/ml for 2 hr @ RT)
Alternate names:
Alpha-1 type II collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA137493 600.00
Check your price
  • CD163 Monoclonal Antibody for Western Blot, IF, IHC (F), IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Human, Non-human primate, Sheep
  • Conjugate Unconjugated
  • Catalog # MA182342
  • Size 200µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Hemoglobin scavenger receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182342 600.00
Check your price
  • Cyclin D1 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA139546
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:25)
Alternate names:
PRAD1 oncogene; BCL-1 oncogene; B-cell lymphoma 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA139546 600.00
Check your price
  • L-Thyroxine T4 Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MIT0501PG
  • Size 1mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIT0501PG 600.00
Check your price
  • Hepatitis B Virus Surface Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MIH9705
  • Size 1mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9705 600.00
Check your price
  • hCG Monoclonal Antibody for Western Blot, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIH9822
  • Size 1mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1:250-1:500)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9822 600.00
Check your price
  • Benzoylecgonine Monoclonal Antibody for ELISA and RIA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MIB0202
  • Size 1mg
Applications
ELISA (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIB0202 600.00
Check your price
  • Estriol Polyclonal Antibody for ELISA
  • Host Sheep
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # PA175135
  • Size 100µL
Applications
ELISA (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175135 600.00
Check your price
  • CD180 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA516836
  • Size 100ug
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
Radioprotective 105 kDa protein; Lymphocyte antigen 64
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516836 600.00
Check your price
  • Collagen I Polyclonal Antibody for IHC (F), IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA185312
  • Size 50µg
Applications
ELISA (1:500-1:3000), Immunohistochemistry (Paraffin, Frozen) (1:10-1:50)
Alternate names:
Alpha-2 type I collagen; Alpha-1 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185312 600.00
Check your price
  • CD31 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Non-human primate, Rat
  • Conjugate FITC
  • Catalog # MA516953
  • Size 500ug
Applications
Flow Cytometry (1:50-1:100)
Alternate names:
PECA1; GPIIA'; EndoCAM
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516953 600.00
Check your price
  • CD11b Monoclonal Antibody for IHC (F), Flow and IP
  • Host Mouse
  • Target Species Bovine, Dog, Cat, Goat, Mustelid, Mink, Pig
  • Conjugate Unconjugated
  • Catalog # MA516604
  • Size 2mL
Applications
Flow Cytometry (Neat), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
CD11B, CR3A, MAC-1, MAC1A, MGC117044, MO1A, SLEB6, Integrin alpha M
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516604 600.00
Check your price
  • RSV Monoclonal Antibody for IF
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA516299
  • Size 2mg
Applications
Immunofluorescence (Assay Dependent)
Alternate names:
human respiratory syncytial virus, RSV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516299 600.00
Check your price
  • HSV Type 1 Monoclonal Antibody for IF
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA516285
  • Size 2mg
Applications
Immunofluorescence (Assay Dependent)
Alternate names:
HSV1, Herpes Simplex I, HSV Type 1, Herpes Simplex 1, HHV, HSV, Herpes Simplex Type 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516285 600.00
Check your price
  • Herpes Simplex Virus Type 1/2 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA516294
  • Size 2mg
Applications
ELISA (Assay Dependent)
Alternate names:
HSV1, Herpes Simplex I, HSV Type 1, Herpes Simplex 1, HHV, HSV, Herpes Simplex Type 1, HSV2, HSV-2, Human herpes virus 2, HHV-2, Herpes Simplex II, HSV Type 2, Herpes Simplex 2, HSV Type II, HSV 1/2, HSV I/II
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516294 600.00
Check your price
  • Helicobacter pylori Monoclonal Antibody for ELISA and LF
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA516301
  • Size 2mg
Applications
ELISA (Assay Dependent), Lateral Flow (Assay Dependent)
Alternate names:
H. pylori, Campylobacter pyloridis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516301 600.00
Check your price
  • Procollagen I C-Peptide Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # BTE0020402
  • Size 200µl
Applications
ELISA (1/10,000)
Alternate names:
Alpha-1 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
BTE0020402 600.00
Check your price
  • NGAL Monoclonal Antibody for Western Blot, IF and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # ABS0432902
  • Size 200µL
Applications
ELISA (1:4000), Immunofluorescence (1:1000), Western Blot (Assay dependent)
Alternate names:
p25; Siderocalin LCN2; SV-40-induced 24P3 protein; Lipocalin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0432902 600.00
Check your price
  • Mycobacterium tuberculosis ESAT-6 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # HYB0760802
  • Size 200µl
Applications
ELISA (1:3,200), Western Blot (Assay dependent)
Alternate names:
M. tuberculosis, myobacter
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB0760802 600.00
Check your price
  • Influenza B NP Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # HYB1160202
  • Size 200µl
Applications
ELISA (Assay dependent), Immunocytochemistry (1:1000), Immunofluorescence (1:1,000), Western Blot (1:20)
Alternate names:
Influenza B Nuclear Protein, Influenza Nuclear Protein, Influenza NP
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1160202 600.00
Check your price
  • Exendin 4 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # ABS0123502
  • Size 200µl
Applications
ELISA (1:4,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0123502 600.00
Check your price
  • Complement Factor H Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB2680102
  • Size 200µl
Applications
ELISA (1:64,000), Western Blot (Assay dependent)
Alternate names:
H factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB2680102 600.00
Check your price
  • Clostridium tetani Tetanus Toxin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # HYB2781002
  • Size 200µl
Applications
ELISA (1:16000)
Alternate names:
C. tetani, pE88_60
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB2781002 600.00
Check your price
  • Apolipoprotein B Monoclonal Antibody for Western Blot, IF, IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB0690202
  • Size 200µl
Applications
ELISA (1/32,000), Western Blot (Assay dependent)
Alternate names:
apo-B
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB0690202 600.00
Check your price
  • Butyrylcholinesterase Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HAH0020102
  • Size 200µg
Applications
ELISA (1/20,000)
Alternate names:
Pseudocholinesterase; Choline esterase II; Butyrylcholine esterase; Acylcholine acylhydrolase
Catalog # Size Price (AUD) Your Price (AUD) Qty
HAH0020102 600.00
Check your price
  • Streptococcus pneumoniae Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA110833
  • Size 200ug
Applications
ELISA (Assay dependent)
Alternate names:
S. pneumoniae, Pneumococcus
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110833 600.00
Check your price
  • P-Glycoprotein Monoclonal Antibody for Western Blot, ICC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Hamster, Human
  • Conjugate Unconjugated
  • Catalog # MA135604
  • Size 1mL
Applications
Flow Cytometry (1:10), Immunocytochemistry (1:20), Immunohistochemistry (Paraffin, Frozen) (1:20), Western Blot (1:10)
Alternate names:
P-glycoprotein 1; ATP-binding cassette sub-family B member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135604 600.00
Check your price
  • Myoglobin Polyclonal Antibody for ELISA
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA175298
  • Size 1mL
Applications
ELISA (Assay Dependent)
Alternate names:
Human myoglobin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175298 600.00
Check your price
  • MRP2 Monoclonal Antibody for Western Blot, ICC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135678
  • Size 1mL
Applications
Flow Cytometry (Assay dependent.), Immunocytochemistry (1:20-1:50), Immunohistochemistry (Paraffin, Frozen) (1:20), Western Blot (1:20-1:50)
Alternate names:
Multidrug resistance-associated protein 2; Canalicular multidrug resistance protein; ATP-binding cassette sub-family C member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135678 600.00
Check your price
  • Mouse anti-Human IgM Secondary Antibody for Western Blot, Flow, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MII0401
  • Size 1mg
Applications
ELISA (1 µg/ml), Flow Cytometry (1 µg/ml), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (0.5-2ug/ml)
Alternate names:
Immunoglobulin M, IgM
Catalog # Size Price (AUD) Your Price (AUD) Qty
MII0401 600.00
Check your price