Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  90  100  110  120  130  140  150  160  170  175  176  177  178  179  180  181  182  183  184  185  186  187  188  189  190  191  192  193  194  195  196  197  198  199  200  201  202  203  204  210  220  230  240  250  260  270  280  300  400  500  600  700  800  900  1000  1100  1622 
5671 - 5700 of 48655 products displayed
  • iNOS Polyclonal Antibody for Western Blot, IF, ICC, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA3030A
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Frozen) (1:500), Immunohistochemistry (Paraffin) (1:200), Western Blot (1:2,000)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS2; NOS type II; Macrophage NOS; Inducible NO synthase; Hepatocyte NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3030A 375.00
Check your price
  • Influenza A M1 Polyclonal Antibody for IF, ICC and ELISA
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA127326
  • Size 100 µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent)
Alternate names:
Influenza Matrix protein M1, Influenza M1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127326 375.00
Check your price
  • IFIT1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA3848
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:2,000)
Alternate names:
Interferon-induced 56 kDa protein; Glucocorticoid-attenuated response gene 16 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3848 375.00
Check your price
  • HSP27 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), IP, ELISA and IM
  • Host Mouse
  • Target Species Dog, Human, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA3015
  • Size 100 µL
Applications
ELISA (Assay dependent), Immunocytochemistry (1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (Paraffin) (1:500), Immunomicroscopy (Assay Dependent), Immunoprecipitation (2 µg), Western Blot (1:500-1:1000)
Alternate names:
Stress-responsive protein 27; Heat shock 27 kDa protein; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein; p25; Heat shock 25 kDa protein; Growth-related 25 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3015 375.00
Check your price
  • HRD1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA146121
  • Size 100 µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Western Blot (1:1000-1:2000)
Alternate names:
Synovial apoptosis inhibitor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146121 375.00
Check your price
  • HP1 alpha Polyclonal Antibody, HRP conjugate for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate HRP
  • Catalog # PA522740
  • Size 100 µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522740 375.00
Check your price
  • HP1 alpha Polyclonal Antibody, DyLight® 650 conjugate for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate DyLight® 650
  • Catalog # PA522738
  • Size 100 µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522738 375.00
Check your price
  • HP1 alpha Polyclonal Antibody, DyLight® 550 conjugate for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate DyLight® 550
  • Catalog # PA522741
  • Size 100 µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522741 375.00
Check your price
  • HP1 alpha Polyclonal Antibody, DyLight® 488 conjugate for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate DyLight® 488
  • Catalog # PA522739
  • Size 100 µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522739 375.00
Check your price
  • HP1 alpha Polyclonal Antibody, Biotin conjugate for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Biotin
  • Catalog # PA522737
  • Size 100 µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Heterochromatin protein 1 homolog alpha; Antigen p25
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522737 375.00
Check your price
  • HMGB1 Polyclonal Antibody, HRP conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Sheep, Rat
  • Conjugate HRP
  • Catalog # PA522722
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:250), Western Blot (0.5-1 µg/ml)
Alternate names:
High mobility group protein 1; Heparin-binding protein p30; Amphoterin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522722 375.00
Check your price
  • HMGB1 Polyclonal Antibody, DyLight® 650 conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Sheep, Rat
  • Conjugate DyLight® 650
  • Catalog # PA522720
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:250), Western Blot (0.5-1 µg/ml)
Alternate names:
High mobility group protein 1; Heparin-binding protein p30; Amphoterin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522720 375.00
Check your price
  • HMGB1 Polyclonal Antibody, DyLight® 550 conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Sheep, Rat
  • Conjugate DyLight® 550
  • Catalog # PA522723
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:250), Western Blot (0.5-1 µg/ml)
Alternate names:
High mobility group protein 1; Heparin-binding protein p30; Amphoterin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522723 375.00
Check your price
  • HMGB1 Polyclonal Antibody, DyLight® 488 conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Sheep, Rat
  • Conjugate DyLight® 488
  • Catalog # PA522721
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:250), Western Blot (0.5-1 µg/ml)
Alternate names:
High mobility group protein 1; Heparin-binding protein p30; Amphoterin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522721 375.00
Check your price
  • HMGB1 Polyclonal Antibody, Biotin conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Sheep, Rat
  • Conjugate Biotin
  • Catalog # PA522719
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:250), Western Blot (0.5-1 µg/ml)
Alternate names:
High mobility group protein 1; Heparin-binding protein p30; Amphoterin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522719 375.00
Check your price
  • Histone H3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA116941
  • Size 100 µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116941 375.00
Check your price
  • HIF3A Polyclonal Antibody, HRP conjugate for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate HRP
  • Catalog # PA522750
  • Size 100 µL
Applications
Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
Neonatal and embryonic PAS protein; Member of PAS protein 7; Inhibitory PAS domain protein; HIF3-alpha-1; Basic-helix-loop-helix-PAS protein MOP7; PAS domain-containing protein 7; Class E basic helix-loop-helix protein 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522750 375.00
Check your price
  • HIF3A Polyclonal Antibody, HRP conjugate for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate HRP
  • Catalog # PA522717
  • Size 100 µL
Applications
Western Blot (1:100-1:200)
Alternate names:
Neonatal and embryonic PAS protein; Member of PAS protein 7; Inhibitory PAS domain protein; HIF3-alpha-1; Basic-helix-loop-helix-PAS protein MOP7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522717 375.00
Check your price
  • HIF3A Polyclonal Antibody, DyLight® 650 conjugate for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 650
  • Catalog # PA522715
  • Size 100 µL
Applications
Western Blot (1:100-1:200)
Alternate names:
Neonatal and embryonic PAS protein; Member of PAS protein 7; Inhibitory PAS domain protein; HIF3-alpha-1; Basic-helix-loop-helix-PAS protein MOP7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522715 375.00
Check your price
  • HIF3A Polyclonal Antibody, DyLight® 550 conjugate for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 550
  • Catalog # PA522751
  • Size 100 µL
Applications
Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
Neonatal and embryonic PAS protein; Member of PAS protein 7; Inhibitory PAS domain protein; HIF3-alpha-1; Basic-helix-loop-helix-PAS protein MOP7; PAS domain-containing protein 7; Class E basic helix-loop-helix protein 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522751 375.00
Check your price
  • HIF3A Polyclonal Antibody, DyLight® 550 conjugate for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 550
  • Catalog # PA522718
  • Size 100 µL
Applications
Western Blot (1:100-1:200)
Alternate names:
Neonatal and embryonic PAS protein; Member of PAS protein 7; Inhibitory PAS domain protein; HIF3-alpha-1; Basic-helix-loop-helix-PAS protein MOP7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522718 375.00
Check your price
  • HIF3A Polyclonal Antibody, DyLight® 488 conjugate for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 488
  • Catalog # PA522749
  • Size 100 µL
Applications
Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
Neonatal and embryonic PAS protein; Member of PAS protein 7; Inhibitory PAS domain protein; HIF3-alpha-1; Basic-helix-loop-helix-PAS protein MOP7; PAS domain-containing protein 7; Class E basic helix-loop-helix protein 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522749 375.00
Check your price
  • HIF3A Polyclonal Antibody, DyLight® 488 conjugate for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 488
  • Catalog # PA522716
  • Size 100 µL
Applications
Western Blot (1:100-1:200)
Alternate names:
Neonatal and embryonic PAS protein; Member of PAS protein 7; Inhibitory PAS domain protein; HIF3-alpha-1; Basic-helix-loop-helix-PAS protein MOP7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522716 375.00
Check your price
  • HIF3A Polyclonal Antibody, Biotin conjugate for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Biotin
  • Catalog # PA522747
  • Size 100 µL
Applications
Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
Neonatal and embryonic PAS protein; Member of PAS protein 7; Inhibitory PAS domain protein; HIF3-alpha-1; Basic-helix-loop-helix-PAS protein MOP7; PAS domain-containing protein 7; Class E basic helix-loop-helix protein 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522747 375.00
Check your price
  • HIF3A Polyclonal Antibody, Biotin conjugate for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Biotin
  • Catalog # PA522714
  • Size 100 µL
Applications
Western Blot (1:100-1:200)
Alternate names:
Neonatal and embryonic PAS protein; Member of PAS protein 7; Inhibitory PAS domain protein; HIF3-alpha-1; Basic-helix-loop-helix-PAS protein MOP7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522714 375.00
Check your price
  • HIF3A Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA116952
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
Neonatal and embryonic PAS protein; Member of PAS protein 7; Inhibitory PAS domain protein; HIF3-alpha-1; Basic-helix-loop-helix-PAS protein MOP7; PAS domain-containing protein 7; Class E basic helix-loop-helix protein 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116952 375.00
Check your price
  • HIF1A Polyclonal Antibody, HRP conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Goat, Hamster, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate HRP
  • Catalog # PA522779
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (1:500-1:2000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522779 375.00
Check your price
  • HIF1A Polyclonal Antibody, DyLight® 650 conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Non-human primate, Rabbit, Rat, Goat, Hamster, Human, Mouse
  • Conjugate DyLight® 650
  • Catalog # PA522777
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (1:500-1:2000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522777 375.00
Check your price
  • HIF1A Polyclonal Antibody, DyLight® 550 conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Goat, Hamster, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate DyLight® 550
  • Catalog # PA522780
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (1:500-1:2000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522780 375.00
Check your price
  • HIF1A Polyclonal Antibody, DyLight® 488 conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Goat, Hamster, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate DyLight® 488
  • Catalog # PA522778
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (1:500-1:2000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522778 375.00
Check your price