Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  90  100  110  120  130  140  150  160  170  180  181  182  183  184  185  186  187  188  189  190  191  192  193  194  195  196  197  198  199  200  201  202  203  204  205  206  207  208  209  210  220  230  240  250  260  270  280  290  300  400  500  600  700  800  900  1000  1100  1611 
5821 - 5850 of 48310 products displayed
  • CCR2 Polyclonal Antibody, DyLight® 650 conjugate for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate DyLight® 650
  • Catalog # PA523039
  • Size 100 µL
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50), Immunofluorescence (1:50), Western Blot (1 µg/ml)
Alternate names:
Monocyte chemoattractant protein 1 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523039 375.00
Check your price
  • CCR2 Polyclonal Antibody, DyLight® 650 conjugate for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 650
  • Catalog # PA523045
  • Size 100 µL
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50), Immunofluorescence (1:50), Western Blot (1 µg/ml)
Alternate names:
Monocyte chemoattractant protein 1 receptor; MCP-1 receptor; JE/FIC receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523045 375.00
Check your price
  • CCR2 Polyclonal Antibody, DyLight® 550 conjugate for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate DyLight® 550
  • Catalog # PA523042
  • Size 100 µL
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50), Immunofluorescence (1:50), Western Blot (1 µg/ml)
Alternate names:
Monocyte chemoattractant protein 1 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523042 375.00
Check your price
  • CCR2 Polyclonal Antibody, DyLight® 550 conjugate for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 550
  • Catalog # PA523048
  • Size 100 µL
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50), Immunofluorescence (1:50), Western Blot (1 µg/ml)
Alternate names:
Monocyte chemoattractant protein 1 receptor; MCP-1 receptor; JE/FIC receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523048 375.00
Check your price
  • CCR2 Polyclonal Antibody, DyLight® 488 conjugate for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate DyLight® 488
  • Catalog # PA523040
  • Size 100 µL
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50), Immunofluorescence (1:50), Western Blot (1 µg/ml)
Alternate names:
Monocyte chemoattractant protein 1 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523040 375.00
Check your price
  • CCR2 Polyclonal Antibody, DyLight® 488 conjugate for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 488
  • Catalog # PA523046
  • Size 100 µL
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50), Immunofluorescence (1:50), Western Blot (1 µg/ml)
Alternate names:
Monocyte chemoattractant protein 1 receptor; MCP-1 receptor; JE/FIC receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523046 375.00
Check your price
  • CCR2 Polyclonal Antibody, Biotin conjugate for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Biotin
  • Catalog # PA523044
  • Size 100 µL
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50), Immunofluorescence (1:50), Western Blot (1 µg/ml)
Alternate names:
Monocyte chemoattractant protein 1 receptor; MCP-1 receptor; JE/FIC receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523044 375.00
Check your price
  • CCR2 Polyclonal Antibody, Biotin conjugate for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Biotin
  • Catalog # PA523038
  • Size 100 µL
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50), Immunofluorescence (1:50), Western Blot (1 µg/ml)
Alternate names:
Monocyte chemoattractant protein 1 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523038 375.00
Check your price
  • Cathepsin E Polyclonal Antibody for Western Blot, IF, ICC, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA316821
  • Size 100 µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (Paraffin) (1:400), Western Blot (1:2000)
Alternate names:
EC 3.4.23.34, Ctse
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316821 375.00
Check your price
  • Catalase Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFPA0060
  • Size 100 µL
Applications
Immunoprecipitation (5 µl), Western Blot (1:2000)
Alternate names:
CTA1
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFPA0060 375.00
Check your price
  • Catalase Monoclonal Antibody for IF, ICC, Flow, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFMA0004
  • Size 100 µL
Applications
ELISA (Assay dependent), Immunoprecipitation (1-2 µl)
Alternate names:
CTA1
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0004 375.00
Check your price
  • Catalase Monoclonal Antibody for IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFMA0003
  • Size 100 µL
Applications
ELISA (Assay dependent), Immunoprecipitation (1-2 µl)
Alternate names:
CTA1
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0003 375.00
Check your price
  • Catalase Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFMA0010
  • Size 100 µL
Applications
Western Blot (1 µg/ml)
Alternate names:
CTA1
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0010 375.00
Check your price
  • Caspase 10 Monoclonal Antibody for Western Blot
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA124702
  • Size 100 µL
Applications
Western Blot (5-10 µg/ml)
Alternate names:
ICE-like apoptotic protease 4; FAS-associated death domain protein interleukin-1B-converting enzyme 2; Apoptotic protease Mch-4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124702 375.00
Check your price
  • Cardiotin Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Dog, Cat, Goat, Hamster, Human, Mouse, Non-human primate, Rabbit, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # OMA106030
  • Size 100 µL
Applications
Immunofluorescence (1:25-1:100), Immunohistochemistry (Paraffin, Frozen) (1:25-100), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
OMA106030 375.00
Check your price
  • Carbonic Anhydrase IX Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P), Flow, ChIP, ELISA and GS
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Plant, Rat
  • Conjugate Unconjugated
  • Catalog # PA116592
  • Size 100 µL
Applications
Gel Shift (Assay-Dependent), Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Immunohistochemistry (Frozen) (1:1000), Immunohistochemistry (Paraffin) (1:1000), Western Blot (1:2000)
Alternate names:
Membrane antigen MN homolog; Carbonic anhydrase IX; Carbonate dehydratase IX; pMW1; Renal cell carcinoma-associated antigen G250; P54/58N; Membrane antigen MN
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116592 375.00
Check your price
  • CaMKII beta Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA141150
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Western Blot (Assay-Dependent)
Alternate names:
CAM2, CAMK2, CAMKB, MGC29528
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141150 375.00
Check your price
  • Calsequestrin Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Dog, Chicken, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3913
  • Size 100 µL
Applications
Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (1:20), Western Blot (1:1,000)
Alternate names:
Laminin-binding protein; Calsequestrin, skeletal muscle isoform; Aspartactin; Calmitine
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3913 375.00
Check your price
  • Calreticulin Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Chicken
  • Target Species Dog, Hamster, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA1903
  • Size 100 µL
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:2,000)
Alternate names:
HACBP; Endoplasmic reticulum resident protein 60; Calregulin; CRP55; Calcium-binding protein 3; CALBP; grp60
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1903 375.00
Check your price
  • Calreticulin Polyclonal Antibody for Western Blot, IF, ICC, IHC, IP, ARRAY, Neu, DB and IM
  • Host Rabbit
  • Target Species Rat, Bovine, Hamster, Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA316862
  • Size 100 µL
Applications
Blocking Assay (Assay-Dependent), Immunocytochemistry (1:50-1:250), Immunofluorescence (1:50-1:250), Immunohistochemistry (1:50), Immunomicroscopy (Assay-Dependent), Immunoprecipitation (Assay-Dependent), Neutralization (Assay-Dependent), Western Blot (1:1000)
Alternate names:
HACBP; Endoplasmic reticulum resident protein 60; Calregulin; CRP55; Calcium-binding protein 3; CALBP; grp60
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316862 375.00
Check your price
  • Calreticulin Polyclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA116924
  • Size 100 µL
Applications
Immunocytochemistry (1:20-1:100), Immunofluorescence (1:20-1:100), Western Blot (1:1000-1:10,000)
Alternate names:
HACBP; Endoplasmic reticulum resident protein 60; Calregulin; CRP55; grp60
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116924 375.00
Check your price
  • Caldesmon Monoclonal Antibody for Western Blot, IF, ICC, ELISA and IDF
  • Host Mouse
  • Target Species Avian, Gerbil, Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA124913
  • Size 100 µL
Applications
ELISA (1:2000), Immunocytochemistry (Assay-dependent), Immunodiffusion (Assay-dependent), Immunofluorescence (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
L-caldesmon
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124913 375.00
Check your price
  • Calcium Sensing Receptor Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA1934A
  • Size 100 µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Western Blot (1:100)
Alternate names:
Parathyroid cell calcium-sensing receptor 1; Parathyroid cell calcium-sensing receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1934A 375.00
Check your price
  • CAF1 p60 Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA116845
  • Size 100 µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunoprecipitation (1:10-1:500), Western Blot (1:100-1:2000)
Alternate names:
M-phase phosphoprotein 7; Chromatin assembly factor I p60 subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116845 375.00
Check your price
  • c-Myc Tag Monoclonal Antibody for Western Blot, IHC, IHC (F), IHC (P), Flow, IP, ChIP and ELISA
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Yeast
  • Conjugate Unconjugated
  • Catalog # MA116637
  • Size 100 µL
Applications
ChIP assay (2 µg/500 µg extract), ELISA (1:100-1:2000), Flow Cytometry (1:200-1:400), Immunohistochemistry (Frozen) (1:100), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (2 µg/mg lysate), Western Blot (1:500-1:1000)
Alternate names:
Transcription factor p64; Proto-oncogene c-Myc; Class E basic helix-loop-helix protein 39
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116637 375.00
Check your price
  • c-Fos Polyclonal Antibody for Western Blot and IHC
  • Host Sheep
  • Target Species Hamster, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA118328
  • Size 100 µL
Applications
Immunohistochemistry (1:500-1:8000), Western Blot (1:500-1:8000)
Alternate names:
Cellular oncogene fos
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118328 375.00
Check your price
  • Bub1b Monoclonal Antibody, HRP conjugate for Western Blot
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate HRP
  • Catalog # MA516038
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Western Blot (1:2000-1:5000)
Alternate names:
Mitotic checkpoint kinase MAD3L; MAD3/BUB1-related protein kinase; Protein SSK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516038 375.00
Check your price
  • Bub1b Monoclonal Antibody, DyLight® 650 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate DyLight® 650
  • Catalog # MA516036
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Western Blot (1:2000-1:5000)
Alternate names:
Mitotic checkpoint kinase MAD3L; MAD3/BUB1-related protein kinase; Protein SSK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516036 375.00
Check your price
  • Bub1b Monoclonal Antibody, DyLight® 550 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate DyLight® 550
  • Catalog # MA516039
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Western Blot (1:2000-1:5000)
Alternate names:
Mitotic checkpoint kinase MAD3L; MAD3/BUB1-related protein kinase; Protein SSK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516039 375.00
Check your price
  • Bub1b Monoclonal Antibody, DyLight® 488 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate DyLight® 488
  • Catalog # MA516037
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Western Blot (1:2000-1:5000)
Alternate names:
Mitotic checkpoint kinase MAD3L; MAD3/BUB1-related protein kinase; Protein SSK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516037 375.00
Check your price