Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  90  100  110  120  130  140  150  160  170  181  182  183  184  185  186  187  188  189  190  191  192  193  194  195  196  197  198  199  200  201  202  203  204  205  206  207  208  209  210  220  230  240  250  260  270  280  290  300  400  500  600  700  800  900  1000  1100  1630 
5851 - 5880 of 48882 products displayed
  • EGFR Monoclonal Antibody for Western Blot, IF, ICC, IHC, IP, ELISA and AP
  • Host Mouse
  • Target Species Dog, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA124678
  • Size 100µL
Applications
Affinity Purification (Assay Dependent), ELISA (1:10,000), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Assay-dependent), Immunoprecipitation (Assay Dependent), Western Blot (1:3000)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124678 375.00
Check your price
  • eGFP Tag Polyclonal Antibody for Western Blot and IHC
  • Host Sheep
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # OSS00005W
  • Size 100µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Enhanced Green Fluorescence Protein, Venus, enhanced GFP
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSS00005W 375.00
Check your price
  • E. coli serotype O/K Polyclonal Antibody, Biotin conjugate for IF and ICC
  • Host Goat
  • Target Species Bacteria
  • Conjugate Biotin
  • Catalog # PA128963
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent)
Alternate names:
Escherichia coli
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128963 375.00
Check your price
  • E. coli serotype O/K Polyclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Goat
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # PA127226
  • Size 100µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Escherichia coli
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127226 375.00
Check your price
  • E-cadherin Monoclonal Antibody for Western Blot, ICC and IHC (F)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA106302
  • Size 100µL
Applications
Immunocytochemistry (Assay dependent), Immunohistochemistry (Frozen) (1:25-1:100), Western Blot (1:50-1:500)
Alternate names:
Uvomorulin; Epithelial cadherin; CAM 120/80
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106302 375.00
Check your price
  • Dynamin 3 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1662
  • Size 100µL
Applications
Western Blot (1:1,000)
Alternate names:
T-dynamin; Dynamin, testicular
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1662 375.00
Check your price
  • Dynamin 2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1661
  • Size 100µL
Applications
Western Blot (1:1,000)
Alternate names:
Dynamin UDNM
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1661 375.00
Check your price
  • Dynamin 1 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1660
  • Size 100µL
Applications
Immunocytochemistry (Assay dependent), Western Blot (1:1,000)
Alternate names:
Dynamin, brain; D100; B-dynamin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1660 375.00
Check your price
  • DUSP2 Polyclonal Antibody for Western Blot, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA3115
  • Size 100µL
Applications
Immunocytochemistry (1:1,000), Immunohistochemistry (Paraffin) (1:500), Western Blot (1:1,000)
Alternate names:
Dual specificity protein phosphatase PAC-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3115 375.00
Check your price
  • DRD1 Monoclonal Antibody for Western Blot, IF, ICC, IHC and IHC (F)
  • Host Mouse
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA146024
  • Size 100µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (Frozen) (1:1000), Western Blot (1:200-1:500)
Alternate names:
Dopamine D1 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146024 375.00
Check your price
  • Dopamine Transporter Polyclonal Antibody for Western Blot and IHC (F)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA14657
  • Size 100µL
Applications
Immunohistochemistry (Frozen) (1:1000), Western Blot (1:1000)
Alternate names:
Solute carrier family 6 member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14657 375.00
Check your price
  • Dopamine Transporter Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA14656
  • Size 100µL
Applications
Immunohistochemistry (1:1000), Western Blot (1:1000)
Alternate names:
Solute carrier family 6 member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14656 375.00
Check your price
  • DOPA Decarboxylase Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Dog, Guinea pig, Human, Sheep, Rabbit, Rat, Bovine
  • Conjugate Unconjugated
  • Catalog # PA14652
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
DOPA decarboxylase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14652 375.00
Check your price
  • DNM1L Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and IP
  • Host Rabbit
  • Target Species Fish, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA116987
  • Size 100µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (Paraffin) (2.5 µg/ml), Immunoprecipitation (Assay-Dependent), Western Blot (1:500)
Alternate names:
Dynamin-like protein; Dynamin-related protein 1; Dynamin-like protein IV; Dynamin-like protein 4; Dynamin family member proline-rich carboxyl-terminal domain less; Dnm1p/Vps1p-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116987 375.00
Check your price
  • DNM1L Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA116986
  • Size 100µL
Applications
Immunocytochemistry (1:50-1:400), Immunofluorescence (1:50-1:400), Immunohistochemistry (Paraffin) (2.5 µg/ml), Western Blot (1:500)
Alternate names:
Dynamin-like protein; Dynamin-related protein 1; Dynamin-like protein IV; Dynamin-like protein 4; Dynamin family member proline-rich carboxyl-terminal domain less; Dnm1p/Vps1p-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116986 375.00
Check your price
  • DISC1 Polyclonal Antibody, HRP conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate HRP
  • Catalog # PA522842
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (2.5-5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
RP4-730B13.1, C1orf136, SCZD9, disrupted in schizophrenia 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522842 375.00
Check your price
  • DISC1 Polyclonal Antibody, HRP conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate HRP
  • Catalog # PA522839
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (2.5-5 µg/ml), Western Blot (1:100-1:2000)
Alternate names:
RP4-730B13.1, C1orf136, SCZD9, disrupted in schizophrenia 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522839 375.00
Check your price
  • DISC1 Polyclonal Antibody, DyLight® 650 conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate DyLight® 650
  • Catalog # PA522841
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (2.5-5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
RP4-730B13.1, C1orf136, SCZD9, disrupted in schizophrenia 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522841 375.00
Check your price
  • DISC1 Polyclonal Antibody, DyLight® 650 conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate DyLight® 650
  • Catalog # PA522837
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (2.5-5 µg/ml), Western Blot (1:100-1:2000)
Alternate names:
RP4-730B13.1, C1orf136, SCZD9, disrupted in schizophrenia 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522837 375.00
Check your price
  • DISC1 Polyclonal Antibody, DyLight® 550 conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate DyLight® 550
  • Catalog # PA522843
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (2.5-5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
RP4-730B13.1, C1orf136, SCZD9, disrupted in schizophrenia 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522843 375.00
Check your price
  • DISC1 Polyclonal Antibody, DyLight® 550 conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate DyLight® 550
  • Catalog # PA522840
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (2.5-5 µg/ml), Western Blot (1:100-1:2000)
Alternate names:
RP4-730B13.1, C1orf136, SCZD9, disrupted in schizophrenia 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522840 375.00
Check your price
  • DISC1 Polyclonal Antibody, DyLight® 488 conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate DyLight® 488
  • Catalog # PA522838
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (2.5-5 µg/ml), Western Blot (1:100-1:2000)
Alternate names:
RP4-730B13.1, C1orf136, SCZD9, disrupted in schizophrenia 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522838 375.00
Check your price
  • DISC1 Polyclonal Antibody, Biotin conjugate for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # PA522836
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (2.5-5 µg/ml), Western Blot (1:100-1:2000)
Alternate names:
RP4-730B13.1, C1orf136, SCZD9, disrupted in schizophrenia 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522836 375.00
Check your price
  • DISC1 Polyclonal Antibody for IF, ICC, IHC, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA146164
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (2.5-5 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146164 375.00
Check your price
  • Di-Methyl-Histone H3 (Lys27) Monoclonal Antibody for Western Blot, IF and ChIP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA511197
  • Size 100µL
Applications
ChIP assay (1:25), Immunofluorescence (1:1600), Western Blot (1:1000)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511197 375.00
Check your price
  • Defensin 5 Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), Flow, ELISA and DB
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA146026
  • Size 100µL
Applications
ELISA (Assay-Dependent), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (1:1000)
Alternate names:
HD5(20-94); Defensin, alpha 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146026 375.00
Check your price
  • Cytokeratin 7 Monoclonal Antibody for Western Blot, ICC, IHC (F) and Flow
  • Host Mouse
  • Target Species Dog, Cat, Goat, Hamster, Human, Mouse, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA106315
  • Size 100µL
Applications
Flow Cytometry (1:100-1:200), Immunocytochemistry (Assay dependent), Immunohistochemistry (Frozen) (1:100-1:200), Western Blot (1:100-1:1000)
Alternate names:
Type-II keratin Kb7; Sarcolectin; Keratin-7; Cytokeratin-7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106315 375.00
Check your price
  • Cytokeratin 19 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA106329
  • Size 100µL
Applications
Flow Cytometry (1:100-1:200), Immunocytochemistry (Assay dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin, Frozen) (1:100-1:200), Western Blot (1:100-1:1000)
Alternate names:
Keratin-19; Cytokeratin-19
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106329 375.00
Check your price
  • Cytokeratin 18 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and Flow
  • Host Mouse
  • Target Species Dog, Chicken, Hamster, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA106326
  • Size 100µL
Applications
Flow Cytometry (Assay dependent), Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Western Blot (1:100-1:1000)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; DreK18; Dorsal aorta proneprin kinesin-1; Cytokeratin endo B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106326 375.00
Check your price
  • Cytokeratin 17 Monoclonal Antibody for Western Blot, ICC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA106325
  • Size 100µL
Applications
Flow Cytometry (1:25-1:200), Immunocytochemistry (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (1:25-1:200), Western Blot (1:100-1:1000)
Alternate names:
Type I keratin Ka17; Keratin-17; Cytokeratin-17; 39.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106325 375.00
Check your price