Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  100  110  120  130  140  150  160  170  180  188  189  190  191  192  193  194  195  196  197  198  199  200  201  202  203  204  205  206  207  208  209  210  211  212  213  214  215  216  217  220  230  240  250  260  270  280  290  300  400  500  600  700  800  900  1000  1100  1200  1630 
6061 - 6090 of 48881 products displayed
  • VEGF Polyclonal Antibody for Western Blot, IHC (F) and Neu
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA547026
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Neutralization (0.2-0.6 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Vascular permeability factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547026 375.00
Check your price
  • VEGF Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P) and ELISA
  • Host Mouse
  • Target Species Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA116629
  • Size 100µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Frozen) (1:20-1:100), Immunohistochemistry (Paraffin) (1:20-1:100), Western Blot (1-2 µg/ml)
Alternate names:
Vascular permeability factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116629 375.00
Check your price
  • VDAC2/VDAC3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Cat, Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA141205
  • Size 100µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Outer mitochondrial membrane protein porin 3; Outer mitochondrial membrane protein porin 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141205 375.00
Check your price
  • VDAC Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA1954A
  • Size 100µg
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (Paraffin) (1:200-1:1000), Western Blot (2 ug/ml)
Alternate names:
Outer mitochondrial membrane protein porin 1; Porin 31HM; Porin 31HL; Plasmalemmal porin; Voltage-dependent anion-selective channel protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1954A 375.00
Check your price
  • VAMP2 Polyclonal Antibody for Western Blot, IF, Flow and IP
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1766
  • Size 100µg
Applications
Immunofluorescence (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (0.3 µg/ml)
Alternate names:
Synaptobrevin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1766 375.00
Check your price
  • VAMP1 Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1765
  • Size 100µg
Applications
Immunoprecipitation (Assay dependent), Western Blot (0.3 µg/ml)
Alternate names:
Synaptobrevin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1765 375.00
Check your price
  • Uromodulin Polyclonal Antibody for Western Blot and IHC (P)
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546959
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (2-15 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
Tamm-Horsfall urinary glycoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546959 375.00
Check your price
  • ULBP3 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547148
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Western Blot (0.1 µg/ml)
Alternate names:
Retinoic acid early transcript 1N; ALCAN-gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547148 375.00
Check your price
  • ULBP2/ULBP5/ULBP6 Polyclonal Antibody for Western Blot, IHC (P) and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547118
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
UL-16 binding protein 5; UL16-binding protein 2; Retinoic acid early transcript 1H; ALCAN-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547118 375.00
Check your price
  • ULBP1 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547131
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Western Blot (0.1 µg/ml)
Alternate names:
UL16-binding protein 1; Retinoic acid early transcript 1I; ALCAN-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547131 375.00
Check your price
  • UGT1A1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547844
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
UDP-glucuronosyltransferase 1A1; UDP-glucuronosyltransferase 1-A; Bilirubin-specific UDPGT isozyme 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547844 375.00
Check your price
  • UCP3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Dog, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1055
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Solute carrier family 25 member 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1055 375.00
Check your price
  • Ubiquilin 1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Hamster, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1759
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
1110046H03Rik, 1810030E05Rik, AU019746, C77538, D13Ertd372e, DA41, Dsk2, Plic-1, XDRP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1759 375.00
Check your price
  • TYROBP Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA112701
  • Size 100µg
Applications
Immunoprecipitation (2-4µg/ml protein lysate), Western Blot (Assay Dependent)
Alternate names:
Killer-activating receptor-associated protein; DNAX-activation protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112701 375.00
Check your price
  • TSLP Polyclonal Antibody for Western Blot, ELISA and Neu
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546975
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Neutralization (0.05-0.25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
thymic stroma-derived lymphopoietin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546975 375.00
Check your price
  • TSG101 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow, IP and ELISA
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA123296
  • Size 100µg
Applications
ELISA (Assay dependent), Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:500-1:3000)
Alternate names:
ESCRT-I complex subunit TSG101
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123296 375.00
Check your price
  • TROP2 Polyclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547074
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Cell surface glycoprotein Trop-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547074 375.00
Check your price
  • TROP2 Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547030
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Pancreatic carcinoma marker protein GA733-1; Membrane component chromosome 1 surface marker 1; Cell surface glycoprotein Trop-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547030 375.00
Check your price
  • TREML2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547471
  • Size 100µg
Applications
Western Blot (0.1 µg/ml)
Alternate names:
Triggering receptor expressed on myeloid cells-like protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547471 375.00
Check your price
  • TREM2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546978
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Triggering receptor expressed on monocytes 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546978 375.00
Check your price
  • TREM1 Polyclonal Antibody for Western Blot, Flow, ELISA and FN
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547112
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Functional Assay (Assay-Dependent), Western Blot (1 µg/ml)
Alternate names:
Triggering receptor expressed on monocytes 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547112 375.00
Check your price
  • TRAP1 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA1010
  • Size 100µg
Applications
Immunocytochemistry (1:250), Immunofluorescence (1:250), Immunohistochemistry (Frozen) (1:20), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (Assay dependent), Western Blot (1:2,000)
Alternate names:
Tumor necrosis factor type 1 receptor-associated protein; TNFR-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1010 375.00
Check your price
  • TRAG3 Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112868
  • Size 100µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
CSAG3B, CT24.2, MGC149851, MGC149852, TRAG3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112868 375.00
Check your price
  • TR2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124354
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Testicular receptor 2; Orphan nuclear receptor TR2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124354 375.00
Check your price
  • TPX2 Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1802
  • Size 100µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (4 ug/mL), Immunohistochemistry (1:200), Immunohistochemistry (Paraffin) (0.5-1.0 ug/ml), Immunoprecipitation (Assay dependent), Western Blot (2 ug/mL)
Alternate names:
Restricted expression proliferation-associated protein 100; Protein fls353; Hepatocellular carcinoma-associated antigen 90; Hepatocellular carcinoma-associated antigen 519; Differentially expressed in cancerous and non-cancerous lung cells 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1802 375.00
Check your price
  • Toxoplasma gondii Monoclonal Antibody, FITC conjugate for Western Blot, IF and ELISA
  • Host Mouse
  • Target Species Protozoa
  • Conjugate FITC
  • Catalog # MA17436
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunofluorescence (1:5), Western Blot (Assay Dependent)
Alternate names:
T. gondii
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17436 375.00
Check your price
  • TNFRSF13C Polyclonal Antibody for Western Blot, Flow and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546904
  • Size 100µg
Applications
Flow Cytometry (0.25 µg/10^6 cells), Neutralization (3-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
BLyS receptor 3; BAFF receptor; B-cell-activating factor receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546904 375.00
Check your price
  • TNFAIP6 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547253
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Tumor necrosis factor alpha-induced protein 6; TNF-stimulated gene 6 protein; Hyaluronate-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547253 375.00
Check your price
  • TNF alpha Monoclonal Antibody for IHC (F), IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126970
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
Tumor necrosis factor ligand superfamily member 2; TNF-alpha; Cachectin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126970 375.00
Check your price
  • TMPRSS15 Polyclonal Antibody for Western Blot and IP
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547162
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Transmembrane protease serine 15; Serine protease 7; Enterokinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547162 375.00
Check your price